Analytical / Chromatography
Air Monitoring
Analytical Reagents
Analytical Standards
Gas Chromatography
HPLC
Microbiology
Microscopy
Sample Preparation
Spectroscopy
Syringes
Titration
TLC
Vials
Videos
HPLC
HYDRANAL
One Component Reagent
Two Component Reagent
Coulomat with Diaphragm
Coulomat no Diaphragm
The Use of Ovens
Water Standard Use
Standardization of Titre
Questions/Comments
SPME
VersaFlash
Events
Literature
Analytix
The Reporter
Reporter Europe
 Coulometric Cell without a Diaphragm
HYDRANAL® Videos
Coulometric Titration with HYDRANAL®-Coulomat Reagents;
Coulometric Cell without a Diaphragm
| View Products | Multimedia Guide on CD |
| Questions / Comments? |
Coulometry without a Diaphragm:
Coulometric cells without a diaphragm consist of only one chamber. The anode and cathode reaction take place in the same electrolyte. The cathode reaction must not produce any side products which could be oxidized. They would consequently give the impression of extra water in the cell.
Hydranal-Coulomat AD/E/AG/AG-H/AK and AG Oven are suitable for this form of coulometry.