|
SPME is a unique sample preparation technique that eliminates
most drawbacks to extracting organics. SPME requires no solvents or complicated
apparatus. It can concentrate volatile and nonvolatile compounds (in liquids
or gaseous samples), for subsequent analysis by GC or HPLC.
SPME offers some important advantages:
- Single Step Extraction - reduces sample preparation time by 70%
- Versatility - various selectivities available, adapts to any GC or
HPLC system, can be automated
- Efficient & Economical - little/no use of solvents, quantitative
method, allows more than 50 extractions per fiber on average
View our SPME video selection below to see how the SPME
technique can meet your sample preparation needs.
Order SPME
products through our online product catalog. |
Vídeo com tradução para Português |
|