Order Center
Biochemicals
Antibiotic Explorer
BioUltra
Amino Acids
Antioxidants
Biological Buffers
Carbohydrates
Chelators
Crystallization
Denaturation
Density Gradient
Detergents/Surfactants
Lumin. Spectroscopy
Molecular Biology
Biomaterials
Detection
Detergent Theory
Enzyme Explorer
Lipids
Microparticles
Pharmacopoeia
PolyAmino Acids
Reference Chart
BioFiles
 Luminescence Spectroscopy Reagents
BioUltra Reagents
Introduction
BioUltra, for Luminescence Quality
References
Product List
To meet highest demands for quality, we offer a selection of BioUltra Luminescence Spectroscopy Reagents.
Introduction
Emission spectra (fluorescence and phosphorescence), and their evaluation with respect to time, are a much more sensitive and specific measurement than absoption spectra, and are particularly useful in trace analysis and in biology and medicine [1,2,3].
During experimentation (separation, purification, analysis), other chemicals are required to support the conditions necessary for investigation. This could lead to the introduction of fluorescent contamination especially if large amounts of reagents are used, with a resultant increase in background fluorescence.
To meet the needs of our customers, a selection of products has been chosen from a wide range of fields of application, and screened to ensure the absence of fluorescent impurities, according to the luminescence test described below. At present, this group consist of several buffers, salts and acids.
BioUltra, for Luminescence Quality
Definition:
In addition to the general BioUltra Reagent tests, this group of product is screened by means of a luminescence test for the absence of fluorescence impurities and thus products can be assessed as suitable for use in techniques using luminescence spectroscopy.
Luminescence Test:
Fluorescent impurities can be detected by subjecting a solution of test material to light excitation between 200 and 620 nm and observing emission in a range between 300 and 700 nm. The product is considered suitable when no impurities are detected.
References
  1. S. Udenfriend, Fluorescence Assay in Biology and Medicine, Academic Press, New York (1962)
  2. S.V. Konev, Fluorescence and Phosphorescence of Proteins and Nucleic Acids, Plenum Press, New York (1963)
  3. Handbook of Analytical Chemistry, McGraw-Hill, New York, pp. 6-176 to 6-196 (1963)
  4. D.J. Kane et al., Biochemistry, 36, 13406 (1997)
Back to Top
Back to BioUltra home
Back to Bioscience home