Click on the column headings to sort products. To re-alphabetize, hit the page refresh button in your web browser.
Cat. No. |
Product Name |
Solvent |
λ Ex Max |
λ Em Max |
Additional Conditions for Fluorescence |
λ Ex Max |
λ Em Max |
| 70684 |
1-(2-Naphthoyl)imidazole |
H2O |
230 |
362 |
Aldite adducts |
234 |
374 |
| 65221 |
1-(4-Methoxyphenyl)-6-phenylhexatriene |
MeOH |
380 |
440 |
|
|
|
| 46123 |
1-(Ethoxycarbonylmethyl)-6-methoxyquinolinium bromide |
H2O |
355 |
460 |
|
|
|
| 34880 |
1,1'-Dibutyl-3,3,3',3'-tetramethylindocarbocyanine iodide |
pH 7.0 |
548 |
598 |
|
|
|
| 36808 |
1,1'-Diethyl-3,3,3',3'-tetramethylindocarbocyanine iodide |
pH 7.0 |
559 |
598 |
|
|
|
| 37250 |
1,1'-Dihexyl-3,3,3',3'-tetramethylindocarbocyanine iodide |
pH 7.0 |
547 |
599 |
|
|
|
| 42364 |
1,1'-Dioctadecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate |
MeOH |
550 |
565 |
|
|
|
| 42368 |
1,1'-Dioctadecyl-3,3,3',3'-tetramethylindodicarbocyanine perchlorate |
MeOH |
642 |
664 |
|
|
|
| 42450 |
1,1'-Dioctyl-3,3,3',3'-tetramethylindocarbocyanine iodide |
pH 7.0 |
546 |
598 |
|
|
|
| 42645 |
1,1'-Dipentyl-3,3,3',3'-tetramethylindocarbocyanine iodide |
pH 7.0 |
548 |
600 |
|
|
|
| 43605 |
1,1'-Dipropyl-3,3,3',3'-tetramethylindocarbocyanine iodide |
pH 7.0 |
548 |
599 |
|
|
|
| 14510 |
1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid |
pH 8.0 |
287 |
363 |
with calcium |
274 |
363 |
| 14513 |
1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid tetrasodium salt |
pH 8.0 |
254 |
363 |
with calcium |
274 |
363 |
| 31535 |
1,2-Diacetylbenzene |
|
|
|
Amine adducts |
355 |
455 |
| 29059 |
1,3-Cyclohexanedione |
|
|
|
Protein adduct |
375 |
450 |
| 36811 |
1,3'-Diethyl-4,2'-quinolylthiacyanine iodide |
DMSO |
470 |
551 |
bound to RNA |
508 |
529 |
| 15090 |
1,4-Bis(2-methylstyryl)benzene |
Cyclohexane |
|
419 |
Heptane |
|
416 |
| 15080 |
1,4-Bis(4-methyl-5-phenyl-2-oxazolyl)benzene |
Dioxane |
|
430 |
|
|
|
| 15150 |
1,4-Bis(5-phenyl-2-oxazolyl)benzene |
Cyclohexane |
358 |
440 |
Toluene |
385 |
418 |
| 33165 |
1,5-Diaminonaphthalene |
H2O:MeOH (1:4) |
330 |
390 |
|
|
|
| 43050 |
1,6-Diphenyl-1,3,5-hexatriene |
MeOH |
350 |
452 |
|
|
|
| 33170 |
1,8-Diaminonaphthalene |
Cyclohexane |
356 |
392 |
|
|
|
| 86181 |
10-(3-Sulfopropyl)acridinium betaine |
H2O |
415 |
488 |
|
|
|
| 44164 |
10-Dodecylacridine Orange Bromide |
MeOH |
493 |
520 |
|
|
|
| 74747 |
10-Octadecylacridine orange bromide |
MeOH |
494 |
519 |
|
|
|
| 82659 |
11-(1-Pyrenesulfonylamino)undecanoic acid |
MeOH |
379 |
398 |
|
|
|
| 72966 |
11-(7-Nitrobenzofurazan-4-ylamino)undecanoic acid |
MeOH |
466 |
530 |
|
|
|
| 39235 |
11-[5-(Dimethylamino)-1-naphthalenesulfonylamino]undecanoic acid |
MeOH |
335 |
519 |
|
|
|
| 72963 |
12-(7-Nitrobenzofurazan-4-ylamino)dodecanoic acid |
MeOH |
467 |
539 |
|
|
|
| 75937 |
12-Oxo-12-(1-pyrenyl)dodecanoic acid |
H2O |
385 |
434 |
|
|
|
| 04510 |
1-Ethylnaphthalene |
DMSO |
260 |
337 |
|
|
|
| 69025 |
1-Methylpyrene |
CHCl3 |
340 |
486 |
|
|
|
| 70930 |
1-Naphthaleneacetic anhydride |
|
|
|
Polyamine adducts |
216 |
360 |
| 70438 |
1-Naphthol |
Borax buffer |
330 |
472 |
|
|
|
| 70440 |
1-Naphthol |
Borax buffer |
330 |
472 |
|
|
|
| 82655 |
1-Pyrenebutyric acid |
MeOH |
341 |
376 |
Reaction product |
342 |
395 |
| 82654 |
1-Pyrenecarboxaldehyde |
MeOH |
345 |
455 |
|
|
|
| 82639 |
1-Pyrenecarboxylic acid |
MeOH |
342 |
382 |
|
|
|
| 82660 |
1-Pyrenedecanoic acid |
MeOH |
341 |
377 |
|
|
|
| 82663 |
1-Pyrenedodecanoic acid |
MeOH |
341 |
377 |
|
|
|
| 82667 |
1-Pyrenehexanoic acid |
MeOH |
341 |
377 |
|
|
|
| 82657 |
1-Pyrenesulfonic acid sodium salt |
MeOH |
344 |
376 |
|
|
|
| 14448 |
2-(4-Biphenylyl)-6-phenylbenzoxazole |
H2O |
|
425 |
CH3CN, MeOH |
|
390 |
| 36804 |
2-(4-Diethylaminostyryl)-1-methylpyridinium iodide |
MeOH |
475 |
589 |
|
|
|
| 39416 |
2-(4-Dimethylaminostyryl)-1-methylquinolinium iodide |
pH 7.0 |
463 |
512 |
|
|
|
| 39422 |
2-(4-Dimethylaminostyryl)-1-octadecylpyridinium perchlorate |
DMSO |
550 |
586 |
|
|
|
| 39414 |
2-(4-Dimethylaminostyryl)-3-methylbenzoxazolium perchlorate |
pH 7.2 |
420 |
555 |
|
|
|
| 39419 |
2-(4-Dimethylaminostyryl)-3-octadecylbenzothiazolium perchlorate |
pH 7.0 |
367 |
603 |
|
|
|
| 63188 |
2-(4-Maleimidophenyl)-6-methylbenzothiazole |
|
|
|
reacted with glutathione |
337 |
393 |
| 69587 |
2'-(4-Methylumbelliferyl)-a-
D
-N-acetylneuraminic acid sodium salt hydrate |
pH 7.0 |
315 |
374 |
Reaction product |
360 |
449 |
| 20526 |
2-(4-tert-Butylphenyl)-5-(4-biphenylyl)-1,3,4-oxadiazole |
EtOH |
|
364 |
|
|
|
| 43131 |
2,5-Diphenyl-1,3,4-oxadiazole |
Toluene |
|
365 |
|
|
|
| 43140 |
2,5-Diphenyloxazole |
Cyclohexane |
303 |
381 |
|
|
|
| 14560 |
2',7'-Bis(2-carboxyethyl)-5(6)-carboxyfluorescein |
pH 7.0 |
482 |
520 |
in alkaline solution |
503 |
528 |
| 14562 |
2',7'-Bis(2-carboxyethyl)-5(6)-carboxyfluorescein tetrakis(acetoxymethyl) ester |
|
|
|
Reaction product |
482 |
528 |
| 35847 |
2',7'-Dichlorofluorescin diacetate |
EtOH |
258 |
|
hydrolyzed |
504 |
529 |
| 39417 |
2-[4-(Dimethylamino)styryl]-1-methylpyridinium iodide |
MeOH |
482 |
504 |
|
|
|
| 57676 |
2-[4-(Iodoacetamido)phenyl]-6-methylbenzothiazole |
CHCl3 |
340 |
396 |
|
|
|
| 06627 |
2-Aminoacridone |
pH 8.0 |
420 |
538 |
|
|
|
| 04520 |
2-Ethylnaphthalene |
EtOH |
300 |
335 |
|
|
|
| 64958 |
2-Methoxy-2,4-diphenyl-3(2H)-furanone |
|
|
|
Amine adducts |
390 |
480 |
| 70450 |
2-Naphthol |
0.1 M NaOH |
320 |
410 |
|
|
|
| 70448 |
2-Naphthol |
0.1 M NaOH |
320 |
410 |
|
|
|
| 54890 |
2-Phenylphenol |
pH 7.9 |
231 |
356 |
|
|
|
| 12779 |
3-(2-Benzothiazolyl)-7-(diethylamino)coumarin |
CH3Cl |
458 |
497 |
|
|
|
| 12781 |
3-(2-Benzothiazolyl)-7-(diethylamino)coumarin-4-carboxylic acid |
MeOH |
450 |
505 |
|
|
|
| 12786 |
3-(2-Benzothiazolyl)-7-octadecyloxycoumarin |
CHCl3 |
390 |
465 |
|
|
|
| 12787 |
3-(2-Benzothiazolyl)umbelliferone |
DMSO |
332 |
408 |
|
|
|
| 12826 |
3-(2-Benzoxazolyl)-4-cyanoumbelliferone |
pH 9.0 |
505 |
577 |
|
|
|
| 12832 |
3-(2-Benzoxazolyl)umbelliferyl acetate |
DMSO |
350 |
485 |
|
|
|
| 12835 |
3-(2-Benzoxazolyl)umbelliferyl octanoate |
DMSO |
350 |
452 |
Reaction product |
410 |
468 |
| 56190 |
3-(4-Hydroxyphenyl)propionic acid |
MeOH |
278 |
|
| 23724 |
3-(5-Chloro-2-benzoxazolyl)-4-cyanoumbelliferone |
pH 4.0 |
418 |
577 |
pH 9.0 |
497 |
577 |
| 30423 |
3-(Dansylamino)phenylboronic acid |
MeOH |
337 |
517 |
Protein adduct |
337 |
490 |
| 34782 |
3,3'-Dibutyloxacarbocyanine iodide |
pH 7.0 |
484 |
532 |
|
|
|
| 34885 |
3,3'-Dibutylthiacarbocyanine iodide |
pH 7.0 |
559 |
604 |
|
|
|
| 36807 |
3,3'-Diethyloxacarbocyanine iodide |
MeOH |
482 |
500 |
|
|
|
| 32446 |
3,3'-Diethyloxadicarbocyanine iodide |
MeOH |
579 |
603 |
|
|
|
| 32448 |
3,3'-Diethyloxatricarbocyanine iodide |
EtOH |
687 |
725 |
|
|
|
| 36809 |
3,3'-Diethylthiacarbocyanine iodide |
pH 7.0 |
379 |
602 |
|
|
|
| 32540 |
3,3'-Diethylthiadicarbocyanine iodide |
MeOH |
651 |
671 |
|
|
|
| 37145 |
3,3'-Diheptyloxacarbocyanine iodide |
MeOH |
482 |
504 |
|
|
|
| 37150 |
3,3'-Diheptylthiacarbocyanine iodide |
MeOH |
562 |
575 |
|
|
|
| 37235 |
3,3'-Dihexyloxacarbocyanine iodide |
MeOH |
484 |
501 |
|
|
|
| 37253 |
3,3'-Dihexylthiacarbocyanine iodide |
pH 7.0 |
547 |
577 |
|
|
|
| 40161 |
3,3-Dimethyl-2-(4-dimethylaminostyryl)-1-octadecylindolium perchlorate |
pH 7.0 |
311 |
390 |
|
|
|
| 42367 |
3,3'-Dioctadecyloxacarbocyanine perchlorate |
MeOH |
484 |
501 |
|
|
|
| 42371 |
3,3'-Dioctadecylthiacarbocyanine perchlorate |
MeOH |
530 |
563 |
|
|
|
| 42455 |
3,3'-Dioctylthiacarbocyanine iodide |
pH 7.0 |
387 |
510 |
|
|
|
| 42600 |
3,3'-Dipentyloxacarbocyanine iodide |
MeOH |
484 |
500 |
|
|
|
| 42647 |
3,3'-Dipentylthiacarbocyanine iodide |
pH 7.0 |
559 |
607 |
|
|
|
| 43580 |
3,3'-Dipropyloxacarbocyanine iodide |
pH 7.0 |
483 |
532 |
|
|
|
| 43607 |
3,3'-Dipropylthiacarbocyanine iodide |
MeOH |
556 |
575 |
|
|
|
| 43608 |
3,3'-Dipropylthiadicarbocyanine iodide |
MeOH |
651 |
675 |
|
|
|
| 41727 |
3,4-Dimethylumbelliferone |
EtOH |
325 |
458 |
|
|
|
| 93282 |
3,6,8-Tris(dimethylaminosulfonyl)-1-pyrenyl phosphate pyridine salt |
H2O |
380 |
470 |
|
|
|
| 31479 |
3,6-Diacetoxyphthalonitrile |
DMSO |
330 |
|
Reaction product |
330 |
445 |
| 43055 |
3-[4-(6-Phenyl-1,3,5-hexatrien-1-yl)phenyl]propionic acid |
DMSO |
350 |
435 |
|
|
|
| 01545 |
3-Acetyl-umbelliferone |
pH 9.0 |
370 |
455 |
|
|
|
| 17631 |
3-Bromomethyl-7-methoxy-1,4-benzoxazin-2-one |
MeOH |
345 |
440 |
|
|
|
| 68163 |
3-Methyl-2-[7-octadecyloxy-3-coumarinyl]benzothiazolium methosulfate |
DMSO |
390 |
510 |
|
|
|
| 74737 |
3-Octadecanoylumbelliferone |
DMSO |
360 |
456 |
|
|
|
| 79279 |
3-Phenylumbelliferone |
pH 3.0 |
338 |
463 |
pH 10.0 |
383 |
472 |
| 79281 |
3-Phenylumbelliferyl phosphate hemipyridine salt |
H2O |
332 |
429 |
|
|
|
| 36806 |
4-(4-Diethylaminostyryl)-1-methylpyridinium iodide |
MeOH |
485 |
607 |
|
|
|
| 42564 |
4-(4-Dipentadecylaminostyryl)-1-methylpyridinium iodide |
pH 7.0 |
470 |
589 |
|
|
|
| 65877 |
4-(6-Methyl-2-benzothiazolyl)phenyl isocyanate |
Chlorobenzene |
350 |
374 |
Amine adduct in CH2Cl2 |
327 |
382 |
| 42359 |
4-(Dioctadecylamino)-4'-nitrostilbene |
MeOH |
440 |
540 |
|
|
|
| 91881 |
4-(Trifluoromethyl)umbelliferone |
MeOH |
385 |
502 |
|
|
|
| 19385 |
4-(Trifluoromethyl)umbelliferyl butyrate |
|
|
|
Reaction product |
385 |
502 |
| 75167 |
4-(Trifluoromethyl)umbelliferyl oleate |
|
|
|
Reaction product |
385 |
502 |
| 91882 |
4-(Trifluoromethyl)umbelliferyl phosphate disodium salt |
|
|
|
Reaction product |
385 |
502 |
| 91880 |
4-(Trifluoromethyl)umbelliferyl-ß-
D
-glucopyranoside |
pH 9.0 |
325 |
413 |
Reaction product |
385 |
502 |
| 91677 |
4,4,4-Trifluoro-3-oxo-2'-butyronaphthone |
MeOH |
333 |
483 |
|
|
|
| 38440 |
4,4'-Diisothiocyanatostilbene-2,2'-disulfonic acid disodium salt hydrate |
H2O |
341 |
415 |
|
|
|
| 66807 |
4,5-Methylenedioxy-1,2-phenylenediamine dihydrochloride |
|
|
|
alpha-Keto acid adducts |
367 |
445 |
| 32670 |
4',6-Diamidino-2-phenylindole dihydrochloride |
H2O |
|
|
bound to DNA |
358 |
461 |
| 17609 |
4-Bromomethyl-6,7-dimethoxycoumarin |
MeOH |
354 |
435 |
Reaction product with acetic acid |
322 |
395 |
| 17623 |
4-Bromomethyl-7-methoxycoumarin |
MeOH |
330 |
|
Reaction product |
322 |
395 |
| 25455 |
4-Chloro-7-nitrobenzofurazan |
MeOH |
336 |
|
Aliphatic amine adducts |
475 |
540 |
| 26375 |
4-Chloro-7-sulfobenzofurazan ammonium salt |
pH 8.5 |
360 |
410 |
Reaction product |
383 |
528 |
| 39250 |
4-Dimethylamino-1-naphthyl isothiocyanate |
MeOH |
343 |
435 |
Amino acid adducts |
240 |
490 |
| 39255 |
4-Dimethylamino-4'-nitrostilbene |
CH2Cl2 |
500 |
700 |
|
|
|
| 47140 |
4-Fluoro-7-nitrobenzofurazan |
EtOH |
470 |
528 |
Amino acid adducts |
470 |
530 |
| 51686 |
4-Heptadecylumbelliferone |
Acidic MeOH |
325 |
385 |
Alkaline MeOH |
367 |
450 |
| 53892 |
4-Hydrazino-7-nitrobenzofurazan |
|
|
|
Aldehyde adduct in chloroform |
445 |
522 |
| 54600 |
4-Hydroxybenzhydrazide |
|
|
|
reacted with 5-hydroxymethyl-furaldehydezinc acetate |
360 |
425 |
| 55150 |
4-Hydroxycoumarin |
MeOH |
|
373 |
|
|
|
| 69580 |
4-Methylumbelliferone |
EtOH |
372 |
445 |
|
|
|
| 51010 |
4-Methylumbelliferyl 4-guanidinobenzoate hydrochloride hydrate |
pH 6.0 |
323 |
464 |
Reaction product |
360 |
449 |
| 69586 |
4-Methylumbelliferyl a-
D
-galactopyranoside |
pH 9.0 |
317 |
374 |
Reaction product |
360 |
449 |
| 69591 |
4-Methylumbelliferyl a-
D
-glucopyranoside |
pH 9.0 |
317 |
374 |
Reaction product |
360 |
449 |
| 69604 |
4-Methylumbelliferyl a-
D
-mannopyranoside |
pH 7.0 |
350 |
375 |
Reaction product |
360 |
449 |
| 69588 |
4-Methylumbelliferyl a-
L
-fucopyranoside |
|
|
|
Reaction product |
360 |
449 |
| 46002 |
4-Methylumbelliferyl acetate |
MeOH |
312 |
|
Reaction product |
360 |
499 |
| 19362 |
4-Methylumbelliferyl butyrate |
MeOH |
312 |
|
Reaction product |
360 |
449 |
| 75230 |
4-Methylumbelliferyl heptanoate |
MeOH |
312 |
|
Reaction product |
360 |
449 |
| 69585 |
4-Methylumbelliferyl N-acetyl-ß-
D
-glucosaminide dihydrate |
pH 10.0 |
317 |
|
Reaction product |
360 |
449 |
| 75164 |
4-Methylumbelliferyl oleate |
MeOH |
312 |
|
Reaction product |
360 |
449 |
| 76163 |
4-Methylumbelliferyl palmitate |
MeOH |
312 |
|
Reaction product |
360 |
449 |
| 69607 |
4-Methylumbelliferyl phosphate |
pH 9.0 |
319 |
384 |
Reaction product |
360 |
449 |
| 69605 |
4-Methylumbelliferyl phosphate disodium salt |
pH 9.0 |
319 |
384 |
Reaction product |
360 |
449 |
| 69590 |
4-Methylumbelliferyl ß-
D
-galactopyranoside |
Glycine buffer |
316 |
376 |
Reaction product |
360 |
449 |
| 69600 |
4-Methylumbelliferyl ß-
D
-glucopyranoside |
pH 9.0 |
316 |
372 |
Reaction product |
360 |
449 |
| 69601 |
4-Methylumbelliferyl ß-
D
-lactopyranoside |
|
|
|
Reaction product |
365 |
445 |
| 69615 |
4-Methylumbelliferyl ß-
D
-N,N',N''-triacetylchitotrioside |
pH 5.4 |
338 |
370 |
|
|
|
| 69610 |
4-Methylumbelliferyl sulfate potassium salt |
pH 10.3 |
334 |
370 |
Reaction product |
360 |
449 |
| 69589 |
4-Methylumbelliferyl-N,N'-diacetyl-ß-
D
-chitobioside monohydrate |
MeOH |
330 |
375 |
|
|
|
| 69602 |
4-Methylumbelliferyl-ß-
D
-glucuronide hydrate |
pH 9.0 |
316 |
375 |
Reaction product |
360 |
449 |
| 73511 |
4-Nitro-4'-(octadecylamino)stilbene |
DMSO |
440 |
|
| 73724 |
4-Nitrophenyl phosphate disodium salt |
pH 7.0 |
310 |
|
Reaction product |
399 |
|
| 73730 |
4-Nitrophenyl phosphate disodium salt hexahydrate |
pH 7.0 |
310 |
|
Reaction product |
399 |
|
| 91879 |
4-Trifluoromethylumbelliferyl ß-
D
-galactopyranoside |
pH 9.0 |
325 |
410 |
Reaction product |
385 |
502 |
| 36565 |
5-(4,6-Dichloro-s-triazin-2-ylamino)fluorescein hydrochloride |
pH 9.0 |
495 |
525 |
|
|
|
| 21882 |
5(6)-Carboxy-2',7'-dichlorofluorescein |
pH 8.0 |
504 |
529 |
|
|
|
| 21884 |
5(6)-Carboxy-2',7'-dichlorofluorescein diacetate |
|
|
|
Reaction product |
470 |
504 |
| 21883 |
5(6)-Carboxyeosin |
pH 8.0 |
519 |
542 |
|
|
|
| 21887 |
5(6)-Carboxyeosin diacetate |
|
|
|
Reaction product |
519 |
542 |
| 21877 |
5(6)-Carboxyfluorescein |
pH > 8.0 |
490 |
520 |
|
|
|
| 21879 |
5(6)-Carboxyfluorescein diacetate |
|
|
|
hydrolyzed |
492 |
517 |
| 21888 |
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester |
DMF |
<300 |
|
hydrolyzed |
492 |
517 |
| 21878 |
5(6)-Carboxyfluorescein N-hydroxysuccinimide ester |
pH 9.0 |
495 |
519 |
|
|
|
| 21933 |
5(6)-Carboxynaphthofluorescein N-succinimidyl ester |
pH 10.0 |
602 |
672 |
|
|
|
| 21953 |
5(6)-Carboxytetramethylrhodamine |
MeOH |
541 |
566 |
|
|
|
| 21955 |
5(6)-Carboxytetramethylrhodamine N-succinimidyl ester |
MeOH |
540 |
565 |
|
|
|
| 21965 |
5(6)-Carboxy-X-rhodamine |
MeOH |
568 |
595 |
|
|
|
| 21966 |
5(6)-Carboxy-X-rhodamine N-succinimidyl ester |
MeOH |
576 |
601 |
|
|
|
| 39225 |
5-(Dimethylamino)-1-naphthalenesulfonamide |
EtOH |
|
580 |
bound to carbon anhydrase |
280 |
470 |
| 57674 |
5-(Iodoacetamido)fluorescein |
pH 9.2 |
493 |
522 |
Mercaptoethanol adduct |
492 |
515 |
| 74735 |
5-(Octadecanoylamino)fluorescein |
CH3OH |
496 |
517 |
|
|
|
| 87639 |
5,10,15,20-Tetrakis(1-methyl-4-pyridinio)porphyrin tetra(p-toluenesulfonate) |
pH 8.0 |
520 |
700 |
|
|
|
| 12340 |
5,6-Benzocoumarin-3-carbonyl chloride |
MeOH |
370 |
438 |
|
|
|
| 86826 |
5-Carboxyfluorescein |
pH 8.0 |
492 |
517 |
|
|
|
| 92846 |
5-Carboxyfluorescein N-succinimidyl ester |
pH 8.0 |
492 |
517 |
|
|
|
| 22575 |
5-Carboxytetramethylrhodamine |
pH 8.0 |
543 |
575 |
|
|
|
| 53048 |
5-Carboxy-tetramethylrhodamine N-succinimidyl ester |
pH 7.0 |
543 |
575 |
|
|
|
| 41969 |
5-Carboxy-X-rhodamine N-succinimidyl ester |
pH 7.0 |
575 |
600 |
|
|
|
| 53047 |
5-Carboxy-X-rhodamine triethylamine salt |
pH 8.0 |
575 |
600 |
|
|
|
| 63184 |
5-Maleimido-eosin |
|
|
|
Mercaptoethanol adduct |
524 |
545 |
| 65320 |
5-Methoxypsoralen |
MeOH |
310 |
480 |
|
|
|
| 72964 |
6-(7-Nitrobenzofurazan-4-ylamino)hexanoic acid |
MeOH |
467 |
539 |
|
|
|
| 89653 |
6-(p-Toluidino)-2-naphthalenesulfonic acid |
MeOH |
318 |
443 |
|
|
|
| 89656 |
6-(p-Toluidino)-2-naphthalenesulfonic acid sodium salt |
MeOH |
318 |
443 |
|
|
|
| 37680 |
6,6'-Dithiodi(2-naphthol) |
|
|
|
Mercaptoethanol adduct |
325 |
369 |
| 36796 |
6,7-Diethoxy-4-methylcoumarin |
MeOH |
340 |
410 |
|
|
|
| 37965 |
6,7-Dihydroxy-4-(trifluoromethyl)coumarin |
EtOH |
370 |
484 |
|
|
|
| 37650 |
6,7-Dihydroxy-4-coumarinylacetic acid |
pH 8.0 |
386 |
465 |
|
|
|
| 37705 |
6,7-Dihydroxy-4-methylcoumarin |
pH 9.0 |
360 |
459 |
|
|
|
| 38828 |
6,7-Dimethoxy-4-(trifluoromethyl)coumarin |
DMSO |
350 |
489 |
|
|
|
| 37920 |
6,8-Dihydroxy-1,3-pyrenedisulfonic acid disodium salt |
pH 4.0 |
405 |
465 |
pH 8.0 |
458 |
500 |
| 46935 |
6-[Fluorescein-5(6)-carboxamido]hexanoic acid |
pH 9.0 |
480 |
525 |
|
|
|
| 46940 |
6-[Fluorescein-5(6)-carboxamido]hexanoic acid N-hydroxysuccinimide ester |
pH 8.2 |
491 |
515 |
|
|
|
| 54115 |
6-Carboxyfluorescein |
pH 8.0 |
492 |
517 |
|
|
|
| 52352 |
6-Carboxyfluorescein N-hydroxysuccinimide ester |
pH 8.0 |
492 |
517 |
|
|
|
| 55222 |
6-Carboxytetramethylrhodamine |
pH 8.0 |
543 |
575 |
|
|
|
| 88997 |
6-Carboxy-tetramethylrhodamine N-succinimidyl ester |
pH 7.0 |
543 |
575 |
|
|
|
| 52472 |
6-Carboxy-X-rhodamine |
pH 8.0 |
575 |
600 |
|
|
|
| 44617 |
6-Carboxy-X-rhodamine N-succinimidyl ester |
pH 7.0 |
575 |
600 |
|
|
|
| 40227 |
6-Dodecanoyl-N,N-dimethyl-2-naphthylamine |
MeOH |
364 |
497 |
|
|
|
| 36803 |
7-(Diethylamino)coumarin-3,4-dicarboxylic acid |
pH 9.0 |
395 |
543 |
|
|
|
| 36798 |
7-(Diethylamino)coumarin-3-carbohydrazide |
MeOH |
420 |
468 |
Carboxylic acid adduct |
415 |
480 |
| 31755 |
7-(Diethylamino)coumarin-3-carbonyl azide |
MeOH |
432 |
480 |
|
|
|
| 36799 |
7-(Diethylamino)coumarin-3-carboxylic acid |
pH 9.0 |
409 |
473 |
|
|
|
| 36801 |
7-(Diethylamino)coumarin-3-carboxylic acid N-succinimidyl ester |
MeOH |
432 |
472 |
|
|
|
| 00871 |
7-Acetoxy-1-methylquinolinium iodide |
|
|
|
Reaction product |
403 |
502 |
| 08445 |
7-Amino-4-methyl-3-coumarinylacetic acid |
MeOH |
350 |
433 |
|
|
|
| 08440 |
7-Amino-4-methylcoumarin |
MeOH |
351 |
430 |
|
|
|
| 46127 |
7-Ethoxy-4-(trifluoromethyl)coumarin |
MeOH |
333 |
415 |
Reaction product |
385 |
502 |
| 46116 |
7-Ethoxy-4-methylcoumarin |
MeOH |
330 |
405 |
Reaction product |
360 |
449 |
| 02644 |
7-Ethoxycoumarin |
pH 7.0 |
323 |
386 |
|
|
|
| 46640 |
7-Fluorobenzofurazan-4-sulfonic acid ammonium salt |
|
|
|
Thiol adduct |
385 |
515 |
| 55158 |
7-Hydroxy-4-coumarinylacetic acid N-succinimidyl ester |
MeOH |
322 |
396 |
|
|
|
| 55627 |
7-Hydroxy-4-methyl-2(1H)-quinolone |
pH 5.0 |
321 |
357 |
pH 9.0 |
351 |
438 |
| 55634 |
7-Hydroxy-4-methyl-3-coumarinylacetic acid |
pH 10.0 |
360 |
445 |
|
|
|
| 55636 |
7-Hydroxy-4-methyl-3-coumarinylacetic acid N-succinimidyl ester |
CHCl3 |
328 |
386 |
|
|
|
| 55155 |
7-Hydroxycoumarin-3-carboxylic acid |
pH 4.0 |
342 |
447 |
pH 9.0 |
386 |
448 |
| 55156 |
7-Hydroxycoumarin-3-carboxylic acid N-succinimidyl ester |
MeOH |
418 |
447 |
|
|
|
| 55157 |
7-Hydroxycoumarin-4-acetic acid |
pH 9.0 |
367 |
455 |
|
|
|
| 56059 |
7-Hydroxy-N-octadecylcoumarin-3-carboxamide |
CHCl3/DMSO |
359 |
406 |
|
|
|
| 65346 |
7-Methoxy-4-(trifluoromethyl)coumarin |
MeOH |
333 |
416 |
|
|
|
| 65087 |
7-Methoxy-4-methylcoumarin |
MeOH |
330 |
405 |
Reaction product |
360 |
449 |
| 64951 |
7-Methoxycoumarin |
CHCl3 |
340 |
389 |
Reaction product |
360 |
449 |
| 64948 |
7-Methoxycoumarin-3-carboxylic acid |
MeOH |
360 |
415 |
|
|
|
| 64949 |
7-Methoxycoumarin-3-carboxylic acid N-succinimidyl ester |
MeOH |
358 |
410 |
|
|
|
| 74759 |
7-Octadecyloxy-3-[3-(3-sulfopropyl)-2-benzothiazolylio]coumarin |
CHCl3 |
390 |
499 |
|
|
|
| 52612 |
8-Acetoxy-N,N,N',N',N'',N''-hexamethylpyrene-1,3,6-trisulfonamide |
MeOH |
394 |
|
| 00879 |
8-Acetoxypyrene-1,3,6-trisulfonic acid trisodium salt |
|
|
|
Reaction product |
460 |
520 |
| 08658 |
8-Aminonaphthalene-1,3,6-trisulfonic acid disodium salt |
pH 7.0 |
356 |
512 |
|
|
|
| 09341 |
8-Aminopyrene-1,3,6-trisulfonic acid trisodium salt |
pH 7.4 |
420 |
500 |
|
|
|
| 10417 |
8-Anilino-1-naphthalenesulfonic acid ammonium salt |
MeOH |
372 |
480 |
Protein adduct |
372 |
480 |
| 20800 |
8-Butyryloxypyrene-1,3,6-trisulfonic acid trisodium salt |
H2O |
384 |
409 |
Reaction product |
460 |
520 |
| 30723 |
8-Decyloxypyrene-1,3,6-trisulfonic acid trisodium salt |
H2O |
404 |
434 |
Quenched with cationic detergents |
|
|
| 44122 |
8-Dodecanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt |
H2O |
370 |
409 |
|
|
|
| 46125 |
8-Ethoxy-N,N,N',N',N'',N''-hexamethylpyrene-1,3,6-trisulfonamide |
|
|
|
Reaction product |
380 |
520 |
| 56360 |
8-Hydroxypyrene-1,3,6-trisulfonic acid tri |