|
E.C. 3.4.21.7
Synonyms: fibrinase; fibrinolysin; actase; serum tryptase; thrombolysin
On this page:
Physical Properties and In vivo processing
Plasminogen:
Plasminogen is the precursor to the fibronolytic protease plasmin as well as angiostatin, an angiogenesis/tumor metastasis inhibitor. Human prostate carcinoma cells release urokinase and free sulfhydryl donors which can produce angiostatin from plasminogen. In vitro, urokinase, tissue plasminogen activator, or streptokinase, in combination with sulfydryl donors, such as N-acetyl-L-cysteine, D-penicillamine, captopril, L-cysteine, or reduced glutathione, produced angiostatin from plasminogen.1
Native-intact human plasminogen is a 291 amino acid glyco-protein with as many as 24 disulfide bonds. Plasminogen contains a single N-linked sialylated biantennary glycan. The two O-glycans possess a Gal β-1-3GalNAc core which is α-2-3 sialylated at the terminal Gal. An additional disialylated form has a second sialic acid residue with an α-2-6 linkage to GalNAc. Mono- and disialylated forms occur at a molar ratio of 80:20 in human plasminogen.2,3
Back to top
Plasmin:
MW: ~86 kDa4 (Human)
When run on SDS-PAGE, bands appeared at 68, 46.2, 23.1 and 12.3 kDa.
Plasmin is a serine endopeptidase of the peptidase S1 family. Plasmin is converted to plasminogen by cleavage between Arg561 and Val562. The resulting activated plasmin consists of two disulfide-linked polypeptide chains. The plasmin heavy chain (MW 60 kDa) is derived from the amino terminal region of plasminogen. The light chain originates from the carboxyl-terminus of plasminogen. In vivo, the MW of the heavy chain can vary from 63KDa to 12KDa depending on the extent of proteolysis to the plasminogen from which it is derived. Plasmin is activated by a variety of proteases including urokinase, tissue plasminogen activator, and streptokinase. During the activation of plasminogen, an autolytic peptide of molecular weight 8,200 is released from the Glu-amino terminus to yield a Lys-amino terminus on the heavy chain. The active site of plasmin is on the light chain.1
Back to top
Specificity and Kinetics
Plasmin exhibits preferential cleavage at the carboxyl side of Lysine and Arginine residues with higher selectivity than trypsin.5 It converts polymerized fibrin into soluble products.
pH Optimum: 8.5
pH 7.5: about 40% of maximal activity, pH 9.5: about 50% of maximal activity6
Temperature Optimum: 37 °C with rapid inactivation at 56 °C6
|
Inhibitors |
| Product # |
Product Name |
Add to Cart | | A1153 |
Aprotinin from bovine lung lyophilized powder, 3-8 TIU/mg solid |
|
| A2504 |
6-Aminocaproic acid ≥99% (titration), powder |
|
| A6191 |
Antipain dihydrochloride from microbial source |
|
| A2221 |
Antithrombin III from human plasma lyophilized powder, ≥95% (SDS-PAGE) |
|
| A7824 |
6-Aminocaproic acid SigmaUltra, ≥99% |
|
| A8456 |
4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride ~97% |
|
| A9141 |
Antithrombin III from bovine plasma lyophilized powder, 200-400 units/mg protein (E1%/280 = 6.5) |
|
| B3906 |
Bdellin from leeches lyophilized powder, >0.1 IU/mg protein (E280) |
|
| C2412 |
C1 Esterase Inhibitor from human plasma aqueous solution, ≥90% (SDS-PAGE) |
|
| D0879 |
Diisopropyl fluorophosphate |
|
| D7910 |
3,4-Dichloroisocoumarin |
|
| E0518 |
C1 Esterase Inhibitor from human plasma aqueous solution, ≥95% (SDS-PAGE) |
|
| E0881 |
Elastatinal microbial, ~50% |
|
| G2417 |
Gabexate mesylate solid |
|
| L2023 |
Leupeptin trifluoroacetate salt ≥90% (HPLC), microbial |
|
| M6159 |
α2-Macroglobulin from human plasma lyophilized powder, ≥98% (SDS-PAGE) |
|
| P7626 |
Phenylmethanesulfonyl fluoride ≥98.5% (GC) |
|
| U4751 |
Urinary Trypsin Inhibitor Fragment ≥95% (HPLC) |
|
| A8849 |
α2-Antiplasmin from human plasma lyophilized powder, ≥5 inhibitor U/mg protein |
|
| Back to top |
|
Substrates |
| Product # |
Product Name |
Add to Cart | | A8171 |
D-Ala-Leu-Lys-7-amido-4-methylcoumarin 95% (HPLC) |
|
| B1136 |
Boc-Val-Leu-Lys-7-amido-4-methylcoumarin |
|
| G8148 |
Gly-Arg-p-nitroanilide dihydrochloride |
|
| I6886 |
D-Ile-Phe-Lys p-nitroanilide ≥95% (HPLC) |
|
| 51015 |
4-Nitrophenyl 4-guanidinobenzoate hydrochloride BioChemika, ≥95.0% (AT) |
|
| T6140 |
N-(p-Tosyl)-Gly-Pro-Lys 4-nitroanilide acetate salt |
|
| 90170 |
Nα-p-Tosyl-L-arginine methyl ester hydrochloride BioChemika, for the determination of trypsin, ≥99.0% (AT) |
|
| V0882 |
D-Val-Leu-Lys 4-nitroanilide dihydrochloride |
|
| Back to top |
Solubility and Solution Stability
Plasmin is readily soluble in water (1 mg/ml). Solutions should be kept on ice and discarded at the end of the day or aliquoted and frozen at a concentration of 1 mg/ml. Frozen solutions will lose approximately 10% activity per month.
Back to top
|
Products |
| Product # |
Product Name |
Add to Cart | | P9156 |
Plasminogen from bovine plasma lyophilized powder, 3-5 units/mg protein |
|
| P5661 |
Plasminogen from human plasma lyophilized powder, ≥2 units/mg protein, suitable for streptokinase determination by clot formation procedure |
|
| P7397 |
Plasminogen from human plasma lyophilized powder, ≥2 units/mg protein, not suitable for streptokinase determination by clot formation procedure |
|
| P1867 |
Plasmin from human plasma lyophilized powder, ≥3 units/mg protein |
|
|
Related Products |
| Product # |
Product Name |
Add to Cart | | F5386 |
Fibrin from human plasma |
|
| S3134 |
Streptokinase from β-hemolytic Streptococcus (Lancefield Group C) lyophilized powder, ≥3,500 units/mg solid |
|
| S8026 |
Streptokinase from β-hemolytic Streptococcus (Lancefield Group C) buffered aqueous solution, ~100,000 units/mg protein |
|
| T5451 |
Tissue Plasminogen Activator from human melanoma cells vial of >5000 IU |
|
| U0633 |
Urokinase from human urine lyophilized powder, ≥500 units/mg protein |
|
| Back to top |
References
- Gatley, S., et al., Proc. Natl. Acad. Sci. U S A, 94, 10868–10872 (1997)
- Marti, T., et al., Eur J Biochem., 173, 57-63 (1988)
- Pirie-Shepherd, S.R., et al., J Biol. Chem.,272, 7408-11 (1997)
- Trends Biochem. Sci., 4, 1 (1979)
- IUBMB Enzyme Nomenclature: www.chem.qmul.ac.uk/iubmb/enzyme/EC3/4/21/7.html
- Greig, B.H.W., et al., Biochim. Biophys. Acta, 67, 658-668 (1963)