Sigma-Aldrich
Sigma - Life Science Logo
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Metabolomics > Enzyme Explorer > Analytical Enzymes > Thrombin Factor IIa
Enzyme Explorer
Thrombin Factor IIa
Thrombin IIa header
On this page:
Related Plasma & Blood Protein Resources:
Specificity
Thrombin is an endolytic serine protease that selectively cleaves the Arg--Gly bonds of fibrinogen to form fibrin and release fibrinopeptides A and B.1,2
The optimal cleavage sites for thrombin have been determined to be 1) A-B-Pro-Arg-||-X-Y where A and B are hydrophobic amino acids and X and Y are nonacidic amino acids and 2) Gly-Arg-||-Gly.2
Thrombin from any mammalian species will clot the fibrinogen of any other mammalian species.3
Thrombin cleaves fibrinogen in 2 ways, but only at arginine sites. The primary cleavage product, fibrinopeptide A is cleaved from fibrinogen after amino acid 16 and sometimes after amino acid 19, while a secondary cleavage product, fibrinopeptide B is produced by cleavage at amino acid 14.4
Thrombin is active in the pH range of 5-10.5
Catalytic optimum is pH 8.3.5
Thrombin precipitates at pH 5 or less.5
Thrombin does not require divalent metal ions or cofactors for activity. However, Na+-dependent allosteric activation of thrombin has been shown to play a role in defining the primary specificity of thrombin to cleave after Arg residues.6 Thromobmodulin serves as a cofactor to thrombin during the activation of protein C.7
back to top
In vivo Processing and Physical Properties
The predominant form of thrombin in vivo is it's zymogen, prothrombin (factor II), which is produced in the liver. The concentration of prothrombin in normal human plasma is ~5-10 mg/dL.8 Prothrombin is a glycoprotein with a glycan content of ~12%.8
Prothrombin is cleaved in vivo by activated factor X releasing the activation peptide and cleaving thrombin into light and heavy chains yielding catalytically active α-thrombin. α-Thrombin is composed of a light chain (A chain)(MW ~ 6,000) and a heavy chain (B chain)(~31,000). These two chains are joined by one disulfide bond.12 The B chain of human thrombin consists of a peptide portion (MW 29,485) and a carbohydrate portion (2334) with N-linked glycosylation at three Asn sites.9,10 Bovine thrombin contains 1.7% glucosamine, 1.8% sialic acid, 0.61% galactose, and 0.95% mannose.11 Thrombin also contains γ-carboxyglutamyl residues. These modified glutamyl residues are the result of carboxylation by a microsomal enzyme, vitamin K-dependent carboxylase. γ-carboxyglutamyl residues are necessary for the calcium-dependent interaction with a negatively charged phospholipid surface, which is essential for the conversion of prothrombin to thrombin.12 In vivo, prothrombin is activated on the surface of a phospholipid membrane that binds the amino terminus of prothrombin along with factors Va and Xa. The activation process starts slowly because factor V itself has to be activated by the initial small amounts of thrombin.
Under certain storage conditions, autolytic digestion of α-thrombin results in the formation of β- and γ-thrombins that lack fibrinolytic activity, but retain some activity against synthetic peptide substrates and protein substrates other than fibrinogen.13 Sigma's thrombin preparations are predominantly the α-thrombin form.
Human thrombin consists of several isozymes with isoelectric points in the range of 6.35-7.6.
For bovine the pI range is 7.05 - 7.114
E1%(280nm) = 18.3 (human)15
E1%(280nm) = 19.5 (bovine)16
Prothrombin_Image
back to top
Measurement of Thrombin Activity
Sigma's thrombin assay procedure is expressed in NIH units obtained by direct comparison to a NIH Thrombin Reference Standard.
The NIH assay procedure uses 0.2 mL of diluted plasma (1:1 with saline) as a substrate and 0.1 mL of thrombin sample (stabilized in a 1% buffered albumin solution at pH 7.35) based on a modification of the method of Biggs.17 Only clotting times in the range of 15-25 seconds are used for determining thrombin concentrations.
Thrombin concentrations in the literature are typically reported in terms of different units of activity.18
Several conventions are used in thrombin literature:
1 IOWA unit= 0.83 NIH unit
1 WHO unit = 0.56 NIH unit
1 NIH unit = 0.324 +/- 0.073 µg
1 NIH unit = 1 USP unit
Thrombin (human and bovine) will catalyze the hydrolysis of several peptide p-nitroanilides, tosyl-arg-nitrobenzyl ester, and a thiobenzyl ester synthetic substrates.19
back to top
Applications
Production of fibrin clot in plasma:
Typicallty one to two units of thrombin will clot one mL of plasma.
Cleavage of Fusion Proteins:
Thrombin can be used for the cleavage of many peptides at the thrombin recognition site using concentrations of 0.5 NIH units thrombin per one nanomole polypeptide in 20 microliters of 50 mM ammonium bicarbonate, pH 8.0.20
Thrombin cleavage of fusion proteins can be carried out at a thrombin to fusion protein ratio of 1:500.21
Fusion proteins may be cleaved in thrombin cleavage buffer consisting of 50 mM Tris, pH 8.0, 150 mM NaCl, 2.5 mM CaCl2 and 0.1% 2-mercaptoethanol. 2 mg of fusion protein was incubated with 4 µg of thrombin for 20 minutes at RT in the cleavage buffer.22
back to top
Products
Human Recombinant Thrombin
Product No. Product Name Add to Cart
T9326 Thrombin from human, recombinant, expressed in HEK 293 cells, ≥2,000 units/mg protein Shopping Cart
Human Thrombin
Product No. Product Name Add to Cart
T1063 Thrombin from human plasma lyophilized powder, >2800 NIH units/mg protein (E1%/280, 18.3) Shopping Cart
T6884 Thrombin from human plasma lyophilized powder, ≥2,000 NIH units/mg protein (E1%/280, 18.3) Shopping Cart
T9010 Thrombin from human plasma lyophilized powder, Suitable for routine use in the thrombin time test Shopping Cart
T7009 Thrombin from human plasma lyophilized powder, ~1,000 NIH units/mg protein (E1%/280, 18.3) Shopping Cart
T9135 Thrombin from human plasma lyophilized powder, Suitable for routine use in the thrombin time test Shopping Cart
T4393 Thrombin from human plasma lyophilized powder, ~1,000 NIH units/mg protein (E1%/280, 18.3), cell culture tested Shopping Cart
T7572 Thrombin from human plasma 50-300 NIH units/mg protein Shopping Cart
Bovine Thrombin
Product No. Product Name Add to Cart
T7513 Thrombin from bovine plasma lyophilized powder, ≥2,000 NIH units/mg protein (E1%/280 = 19.5) Shopping Cart
T9549 Thrombin from bovine plasma powder, cell culture tested, 1,500-2,500 NIH units/mg protein (E1%/280 = 19.5) Shopping Cart
T9681 Thrombin from bovine plasma lyophilized powder, ≥700 NIH units/mg protein (Bradford) Shopping Cart
T6634 Thrombin from bovine plasma lyophilized powder, 600-1,500 NIH units/mg protein (biuret) Shopping Cart
T4265 Thrombin from bovine plasma lyophilized powder, 175-350 NIH units/mg protein (biuret) Shopping Cart
T9000 Thrombin from bovine plasma buffered aqueous solution, ≥125 NIH units/mg protein (biuret) Shopping Cart
T6200 Thrombin from bovine plasma lyophilized powder, 40-300 NIH units/mg protein (biuret) Shopping Cart
T3399 Thrombin from bovine plasma lyophilized powder, 50-150 NIH units/mg protein (biuret) Shopping Cart
Murine Thrombin
Product No. Product Name Add to Cart
T8397 Thrombin from murine plasma lyophilized powder, ~1,000 NIH units/mg protein (biuret) Shopping Cart
Rabbit Thrombin
Product No. Product Name Add to Cart
T4897 Thrombin from rabbit plasma Shopping Cart
Rat Thrombin
Product No. Product Name Add to Cart
T5772 Thrombin from rat plasma lyophilized powder, ≥1,000 units/mg protein (biuret) Shopping Cart
back to top
Substrates
Natural Substrates
Fibrinogen, Factor V,23 Factor VIII,23 Factor XIII,23 Thrombospondin,24 Prothrombin,12 and Protein C.25
Synthetic Substrates
N-Benzoyl-Phe-Val-Arg-p-nitroanilide, N-Benzoyl-Phe-Val-Arg 4-methoxy-b-naphthylamide, N-Benzoyl-Phe-Val-Arg-p-nitroanilide, Boc-β-benzyl-Asp-Pro-Arg-7-amido-4-methylcoumarin, Boc-Val-Pro-Arg-7-amido-4-methylcoumarin, Sar-Pro-Arg p-nitroanilide, Thrombin generation chromogenic, N-p-Tosyl-Gly-Pro-Arg 7-amido-4-methylcoumarin, N-(p-Tosyl)-Gly-Pro-Arg p-nitroanilide.
Inhibitors
Diisopropylfluorophosphate,26 phenylmethylsulfonylfluoride, 26 AEBSF, hirudin,26 tetranitromethane,26 proflavine,26 antithrombin III,26 a1-antitrypsin26 , a1-antiplasmin,26 gabexate mesylate,27 antipain,28 and tosyl-L-lysinechloromethylketone.29
back to top
References
  1. Enzyme Nomenclature: EC 3.4.21.5
  2. Chang, J.Y., Eur. J. Biochem., 151, 217?224 (1985).
  3. The Plasma Proteins, 2nd ed., 2, Putnam, F. W., ed, p. 148.
  4. Machovich, R., The Thrombin, 1, 63-66 (1984)
  5. Machovich, R., The Thrombin, 1, 111 (1984)
  6. Prasad, S., J. Biol. Chem. 279, 10103-10108 (2004)
  7. Kisiel, W., Human plasma protein C: isolation, characterization, and mechanism of activation by alpha-thrombin. J. Clin. Invest. 64, 761-769, (1979)
  8. The Plasma Proteins, 2nd ed., 2, Putnam, F. W., ed: Table 2. See also: The Enzyme Explorer: Plasma and Blood Protein Resource
  9. Qian, W.J., et al., J. Proteome Res., 4, 2070-2080 (2005).
  10. Nilsson, B., et al., Arch. Biochem. Biophys., 224, 127-133 (1983)
  11. Boyer, P.D., The Enzymes, Academic Press (New York), 3rd ed., Vol. III, p. 277-321 (1971).
  12. Expasy/SwissProt: P00743
  13. Boissel, J.P., et al., J. Biol. Chem., 259, 5691-5697 1984).
  14. Righetti, P.G., and Tudor, G., Isoelectric points and molecular weights of proteins, a new table. Journal of Chromatography, 220, 115-194 (1981).
  15. Butkowski, R.J. et al., J. Biol. Chem., 252, 4942 (1977).
  16. Winzor, D. J. and Scheraga, H. A., Arch. Biochem. Biophys. 104, 202-207 (1964)
  17. Human Blood Coagulation, Haemostasis and Thrombosis, 2nd ed., R. Biggs, ed., p. 722 (1976).
  18. The Handbook of Synthetic Substrates, Hemker, H. C., Martinus Nijhoff publisher (1983).
  19. Lottenberg, R., et al., Assay of Coagulation Proteases Using Peptide Chromogenic and Fluorogenic Substrates. Meth. Enzymol., 80-C, 341-361 (1981).
  20. Chang, Y., Thrombin specificity. Requirement for apolar amino acids adjacent to the thrombin cleavage site of polypeptide substrate. Eur. J. Biochem., 151(2), 217-224 (1985).
  21. Hakes, D.J. and Dixon, J.E., Anal. Biochem., 202, 293 (1992).
  22. Gaun, KL and Dixon, JE,, Anal. Biochem., 192, 262, 1991
  23. De Cristofaro, R. and De Candia, E., J. Thromb. Thrombolysis, 15, 151-163 (2003)
  24. Sherwood, J.A., Mol. Biochem. Parisitol., 40, 173-181 (1990)
  25. Berg, D.T., et al., Science, 273, 1389-1391 (1996)
  26. Lundblad, R.L. et al., Methods Enzymol., 45, 156 (1976)
  27. Matsuoka, S., et al., JP. J. Pharmacol., 51, 455-463 (1989)
  28. Wimen, B., Meth. Enzymol., 80, 395-408 (1981)
  29. Magnusson, S. The Enzymes, 3rd ed., III, pp. 277-321, Boyer, P.D., ed., Academic Press (1971)
back to top