Sigma-Aldrich
Sigma - Life Science Logo
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Enzyme Explorer
Trypzean
Sigma® Product # T3568
Recombinant, Animal-Free Bovine Trypsin, Expressed in Corn
Trypzean™ eliminates the introduction of animal source contaminants found in traditional bovine and porcine trypsins.
Specifications:
Appearance: White Powder
Protein Content: ³90% [E1% A280 nm = 14.3]
Specific Activity: Not less than 3650 USP units/mg solid
Molecular Weight: 23,000–27,000 Da by SDS PAGE
Solubility: 500,000 USP Trypsin Units are soluble in 10 mL of water and in 10 mL of saline TS
Features and Benefits:
  • High Purity – Eliminates contaminating activities and increases specificity.
  • Free of animal contaminants – Eliminates bovine pathogen concerns.
  • Comparable kinetics to native trypsin – Simplifies transition from native source trypsin products.
  • Suitability for dissociation of adherent cell lines at several fold lower concentrations than traditional native trypsins – Reduces amount of trypsin required.
  • Inhibition by soybean trypsin inhibitor is comparable to native trypsins on a weight to weight basis – Reduces amount of trypsin inhibitor required.
Kinetics of TrypZean™:
Kinetics of TrypZean
Concentration dependant rates of hydrolysis of BAPNA are essentially identical for TrypZean™ and bovine trypsin.
Availability:
Research Sizes:  10mg and 100mg
Bulk:  Contact Sigma-Aldrich® Fine Chemicals
back to top
back to Enzyme Explorer