Order Center
Biochemicals
Antibiotic Explorer
BioUltra
Biomaterials
Detection
Detergent Theory
Enzyme Explorer
BioUltra Proteins
Product Indexes
Cell Signaling Enzymes
Analytical Enzymes
Enzyme Reagents
Key Resources
Lipids
Microparticles
Pharmacopoeia
PolyAmino Acids
Reference Chart
BioFiles
 Diagnostic & Analytical Enzymes
Enzyme Explorer
Product Name Product # 
Alcohol Dehydrogenase from Saccharomyces cerevisiae, suitable for recycling micro-assay of b-NAD and b-NADH, Lyophilized powder, Solid with buffer salts, minimum 300 units/mg protein A3263
Alcohol Dehydrogenase from Saccharomyces cerevisiae, suitable for conventional determination of b-NAD, b-NADH, ethanol and acetaldehyde, Lyophilized powder, Solution A7011
Alcohol Dehydrogenase equine liver, Lyophilized powder, ³0.5 units/mg protein (TCA/Biuret) A9589
Alcohol Dehydrogenase from Saccharomyces cerevisiae, BioChemika, powder, off-white, ³250 U/mg 05635
Alcohol Dehydrogenase from Saccharomyces cerevisiae, BioChemika, lyophilized, powder, ~300 U/mg 05640
Alcohol Dehydrogenase equine liver, BioChemika, suspension in 20 mM potassium phosphate buffer, pH 7; ethanol 10%, ampoule, 10% as ethanol, ~30 U/ml 05645
Alcohol Dehydrogenase equine liver, BioChemika, crystalline suspension in 20 mM potassium phosphate buffer, ~3 U/mg protein (~10 mg/ml) 05646
Alcohol Dehydrogenase equine liver, BioChemika, lyophilisate, slightly beige, ³0.25 U/mg 17233
Alcohol Dehydrogenase from Candida boidinii, BioChemika, powder, light brown, ³0.4 U/mg 91031
Alcohol Dehydrogenase Thermoanaerobium brockii, Lyophilized powder, 5-15 units/mg protein A8435
Alcohol dehydrogenase, NADP+ dependent Thermoanaerobium brockii, Lyophilized powder, 30-90 units/mg protein A9287
Alcohol Dehydrogenase from Lactobacillus kefir, BioChemika, powder, beige, ~0.4 U/mg 05643
Alcohol Dehydrogenase Thermoanaerobium brockii, BioChemika, powder, white, ~35 U/mg 05655
Alcohol Oxidase Candida boidinii, Lyophilized powder, 5-15 units/mg protein A6941
Alcohol Oxidase from Hansenula sp., Vacuum-dried powder, 20-40 units/mg protein (Bradford) A0438
Alcohol Oxidase solution from Pichia pastoris, Buffered aqueous solution, 10-40 units/mg protein (Biuret) A2404
Alcohol Oxidase solution from Pichia pastoris, BioChemika, solution, red to deep red, ³400 U/ml 92705
(S)-Aromatic Alcohol Dehydrogenase, NADP+ dependent from Thermoanaerobium sp., Lyophilized powder, 1-5 units/mg solid A9685
Bilirubin Oxidase from Myrothecium verrucaria, Lyophilized powder, 15-65 units/mg protein B0390
Catalase preparation from Aspergillus niger, Ammonium sulfate suspension, 4,000-8,000 units/mg protein C3515
Catalase from bison liver, 2,000-5,000 units/mg protein C9447
Catalase bovine liver, Aqueous solution, minimum 40,000 units/mg protein (E1%/405) C3155
Catalase bovine liver, 2,000-5,000 units/mg protein C9322
Catalase from canine liver, Ammonium sulfate suspension, minimum 900 units/mg protein C4138
Catalase from human erythrocytes, minimum 90% (SDS-PAGE), Buffered aqueous solution, >30,000 units/mg protein C3556
Catalase from murine liver, Ammonium sulfate suspension, minimum 900 units/mg protein C8531
Catalase bovine liver, BioChemika, for technical purposes, in 30% glycerol, ³200000 U/ml 60640
Catalase bovine liver, BioChemika, crystalline suspension in water, pH ~6, ~65000 U/mg protein (~20 mg/ml) 60630
Catalase bovine liver, BioChemika, shipped in dry ice, powder, dark green, 10000-25000 U/mg 60629
Catalase bovine liver, BioChemika, lyophilized, powder, brown, ~2500 U/mg 60635
Catalase bovine liver, BioChemika, powder, dark-brown, ~1300 U/mg 60632
Catalase from microorganisms, BioChemika, lyophilized, powder, brown, ~1700 U/mg 60633
Catalase Aspergillus niger, BioChemika, suspension in 3.2 M ammonium sulfate solution, pH6, suspension, dark green-brown, ~70000 U/ml 60628
Catalase Aspergillus niger, BioChemika, ~170 U/mg 60631
Chloramphenicol Acetyltransferase from Escherichia coli, Buffered aqueous glycerol solution C8413
Chloramphenicol Acetyltransferase from Escherichia coli, Lyophilized powder, 50,000-150,000 units/mg protein (Lowry) C2900
Cholesterol Esterase bovine pancreas, Lyophilized powder, 800-1,600 units/g protein C3766
Cholesterol Esterase bovine pancreas, Lyophilized powder, minimum 20,000 units/g protein C5921
Cholesterol Esterase from porcine pancreas, Lyophilized powder, minimum 15,000 units/g protein C9464
Cholesterol Esterase from Pseudomonas sp., Lyophilized powder, minimum 200,000 units/g protein C1403
Cholesterol Esterase from Pseudomonas sp., Lyophilized powder, minimum 10,000 units/g protein C9281
Cholesterol Esterase from porcine pancreas, BioChemika, lyophilized, powder, white, ~35 U/mg 26745
Cholesterol Oxidase microbial, recombinant, expressed in Escherichia coli, Lyophilized powder C1235
Cholesterol Oxidase from Cellulomonas sp., Buffered aqueous solution, 20-60 units/mg protein (Biuret) C5421
Cholesterol Oxidase from Pseudomonas fluorescens, Lyophilized powder, 10-50 units/mg protein C7149
Cholesterol Oxidase from Streptomyces sp., Lyophilized powder, minimum 25 units/mg protein C8649
Cholesterol Oxidase from Pseudomonas sp., BioChemika, powder, light yellow, >1 U/mg 26747
Butyrylcholinesterase from human serum, Lyophilized powder, minimum 3 units/mg C9971
Creatinase from Actinobacillus sp., Lyophilized powder, 20-40 units/mg protein C2409
Creatinase from Flavobacterium sp., Lyophilized powder, 10-20 units/mg protein C7024
Creatinase Pseudomonas sp., recombinant, expressed in Escherichia coli, Lyophilized powder, 10-15 units/mg protein C3921
Creatinase from Flavobacterium sp., BioChemika, lyophilized, white, ~10 U/mg 27905
Creatine Kinase BB Fraction human, >90% (SDS-PAGE), Liquid, recombinant, expressed in Pichia pastoris C9983
Creatine Kinase MB Fraction from human heart, approx. 700 I.U./mL, >90% (SDS-PAGE), Liquid C0984
Creatine Kinase MM Fraction from human heart, pH 7.5, recombinant, expressed in Pichia pastoris, >90% (SDS-PAGE), Lyophilized powder C9858
Creatine Kinase, Type 2 human, minimum 90% (SDS-PAGE), Liquid, approx. 445 units/mg C9608
Creatine Phosphokinase from bovine heart, Type III, Salt-free, lyophilized powder, 150-300 units/mg protein C7886
Creatine Phosphokinase from rabbit brain, Type IV, Lyophilized powder, 50-150 units/mg protein C6638
Creatine Phosphokinase from rabbit muscle, BioChemika, lyophilized, powder, white, ~14 U/mg 27957
Creatine Phosphokinase from rabbit muscle, BioChemika, lyophilized, powder, ~20 U/mg 27960
Creatininase from Flavobacterium sp., suitable for enzymatic determination of creatinine., Lyophilized powder, 150-400 units/mg solid, pH 6.5, 37°C C7399
Creatininase Pseudomonas sp., suitable for enzymatic determination of creatinine., Lyophilized powder, 100-300 units/mg protein, pH 8.0, 25°C C3172
Galactose Oxidase from Dactylium dendroides, Lyophilized powder, minimum 3,000 units/g solid G7400
b-1®(3,6)-Galactosidase, positionally specific, recombinant, expressed in Escherichia coli, Buffered aqueous solution G0288
b-1®4-Galactosidase, positionally specific, recombinant, expressed in Escherichia coli, Buffered aqueous solution G0413
Glucose Oxidase Aspergillus niger, 2,000-10,000 units/g solid (without added oxygen) G6766
Glucose Oxidase Aspergillus niger G7146
Glucose Oxidase Aspergillus niger, Buffered aqueous solution, minimum 200 units/mg protein (Biuret) G9010
Glucose Oxidase Aspergillus niger, Type II, 15,000-25,000 units/g solid (without added oxygen) G6125
Glucose Oxidase Aspergillus niger, Type II-S, 25,000-50,000 units/g solid (without added oxygen) G6641
Glucose Oxidase Aspergillus niger, Type VII, Lyophilized powder, 100,000-200,000 units/g solid (without added oxygen) G2133
Glucose Oxidase Aspergillus niger, Type VII-S, Lyophilized powder, 100,000-250,000 units/g solid (without added oxygen) G7016
Glucose Oxidase Aspergillus niger, Type V-S, approx. 1,000 units/mL (without added oxygen), Buffered aqueous solution G6891
Glucose Oxidase Aspergillus niger, Type X-S, Lyophilized powder, 100,000-250,000 units/g solid (without added oxygen) G7141
Glucose Oxidase Aspergillus niger, BioChemika, solution (clear), yellow, ~1000 U/ml 49177
Glucose Oxidase Aspergillus niger, BioChemika, lyophilized, powder, ~200 U/mg 49180
Glucose Oxidase Aspergillus niger, BioChemika, powder, yellow, ~150 U/mg 49182
Glucose Oxidase Aspergillus niger, BioChemika, powder, yellow, ~130 U/mg 49181
Glucose Oxidase Aspergillus niger, BioChemika, lyophilized, powder, ~25 U/mg 49178
Glucose oxidase/peroxidase reagent G3660
b-Glucosidase from almonds, Lyophilized powder, minimum 2 units/mg solid G0395
b-Glucosidase from almonds, BioChemika, lyophilized, powder, ³6 U/mg 49290
b-Glucuronidase from Helix pomatia, Type H-3AF, Crude solution G0762
b-Glucuronidase Escherichia coli G8271
b-Glucuronidase Escherichia coli G8396
b-Glucuronidase from bovine liver, Type B-10 G0501
b-Glucuronidase from bovine liver, Type B-3 G0376
b-Glucuronidase from bovine liver, Type B-1 G0251
b-Glucuronidase from Helix pomatia, Type H-1 G0751
b-Glucuronidase from Helix pomatia, Type H-2 G0876
b-Glucuronidase from Helix pomatia, Type H-3 G8885
b-Glucuronidase from Helix pomatia, Type H-5 G1512
b-Glucuronidase Helix aspersa (garden snail), Type HA-4 G4259
b-Glucuronidase from Helix pomatia, Type HP-2 G7017
b-Glucuronidase from Helix pomatia, Type HP-2S G7770
b-Glucuronidase Escherichia coli, Type IX-A, Lyophilized powder, 1,000,000-5,000,000 units/g protein (30 min assay) G7396
b-Glucuronidase from Patella vulgata (keyhole limpet), Type L-II, Lyophilized powder, 1,000,000-3,000,000 units/g solid G8132
b-Glucuronidase Escherichia coli, Type VII-A, Lyophilized powder, 5,000,000-20,000,000 units/g protein, pH 6.8 (30 min. assay) G7646
b-Glucuronidase Escherichia coli, Type VIII-A, Aqueous glycerol solution, 5,000,000-20,000,000 units/g protein, pH 6.8 (Biuret, 30 min assay) G7771
b-Glucuronidase Escherichia coli, Type X-A, Lyophilized powder, 20,000,000-60,000,000 units/g protein, pH 6.8 (30 min assay) G7896
b-Glucuronidase Escherichia coli, Aqueous glycerol solution, 20,000,000-60,000,000 units/g protein, pH 6.8 (Biuret) G8162
b-Glucuronidase from Helix pomatia, BioChemika, solution (clear), dark brown, ~5 U/ml 49294
b-Glucuronidase from Helix pomatia, BioChemika, powder, brown, 5-10 U/g 49295
b-Glucuronidase from bovine liver, BioChemika, powder, yellow-brown, ~0.2 U/mg 49309
b-Glucuronidase Escherichia coli, BioChemika, lyophilized, 25%, powder, yellow, 3-15 U/mg 49313
b-Glucuronidase Escherichia coli, BioChemika, powder, yellow-brown, ~1.5 U/mg 49312
b-Glucuronidase from E. coli K12, BioChemika, in 50% (v/v) glycerol, ~100 U/ml 49315
Glutamic-oxalacetic transaminase from porcine heart, Type I, Ammonium sulfate suspension, 200-500 units/mg protein G2751
Glutamic-Oxalacetic Transaminase from porcine heart, Type II-A, Lyophilized powder, 100-400 units/mg protein G7005
Glutamic-Oxalacetic Transaminase from porcine heart, BioChemika, suspension, yellow, ~1400 U/ml 49396
Glutamic-Pyruvic Transaminase from porcine heart, Lyophilized powder, 80-160 units/mg protein G8255
Glutamic-Pyruvic Transaminase from porcine heart, Ammonium sulfate suspension, minimum 75 units/mg protein G9880
Glutamic-Pyruvic Transaminase from porcine heart, BioChemika, in 1.8 M ammonium sulfate solution, pH ~6.0, suspension (aqueous), greenish-yellow, ³100 U/mg protein 49400
Glutathione Peroxidase from bovine erythrocytes, Lyophilized powder, 300-700 units/mg protein G6137
Glutathione Peroxidase from bovine erythrocytes, BioChemika, lyophilized, shipped in dry ice, powder, white, ~100 U/mg 49753
3a-Hydroxysteroid Dehydrogenase preparation from Pseudomonas testosteroni, Lyophilized powder, 20-50 units/mg protein H1506
12a-Hydroxysteroid Dehydrogenase from Bacillus sphaericus, Lyophilized powder, 150-350 units/mg protein (Lowry) H2265
Hydroxysteroid Dehydrogenase from Pseudomonas testosteroni ATCC 11996, minimum 0.5 units/mg protein (using either androsterone or testosterone as substrate (a- and b-activity, respectively)) H8879
Hydroxysteroid Dehydrogenase from Pseudomonas testosteroni ATCC 11996, a-Enzyme 0.5-1.5 units/mg protein (using androsterone as substrate), b-Enzyme 3-5 units/mg protein (using testosterone as substrate) H9004
b-Lactamase from Enterobacter cloacae, BioChemika, purum, ³97.0% (HPLC), powder, light beige, 0.2-0.6 U/mg b 61304
Penicillinase from Bacillus cereus, BioChemika, lyophilized, powder, white, ~13 U/mg 61305
Lactate Oxidase from Pediococcus sp., Lyophilized powder, 20-40 units/mg solid L0638
D-Lactic Dehydrogenase solution from Lactobacillus leichmanii, Ammonium sulfate suspension, 250-500 units/mg protein (Biuret) L2011
L-Lactic Dehydrogenase Bacillus stearothermophilus, recombinant, expressed in Escherichia coli, Lyophilized powder, minimum 200 units/mg protein, pH 6.0, 30°C (Lowry) L3151
D-Lactic Dehydrogenase Leuconostoc mesenteroides, Ammonium sulfate suspension, 1,000-3,000 units/mg protein (Biuret) L2395
L-Lactic Dehydrogenase solution porcine heart, Ammonium sulfate suspension, minimum 400 units/mg protein (Biuret) L7525
L-Lactic Dehydrogenase solution from rabbit muscle, Type I, Ammonium sulfate suspension, 40-100 units/mg protein L2375
L-Lactic Dehydrogenase solution from rabbit muscle, Type II, Ammonium sulfate suspension, 800-1,200 units/mg protein L2500
L-Lactic Dehydrogenase solution bovine heart, Type III, Ammonium sulfate suspension, 500-700 units/mg protein L2625
L-Lactic Dehydrogenase (cytochrome) from baker's yeast (S. cerevisiae), Type IV-SS, Ammonium sulfate suspension, 0.1-0.6 units/mg protein (Biuret) L4506
L-Lactic Dehydrogenase solution bovine heart, Type IX, Ammonium sulfate suspension, minimum 400 units/mg protein L0377
L-Lactic Dehydrogenase from chicken heart, Type VIII, Ammonium sulfate suspension, 500-800 units/mg protein L9126
L-Lactic Dehydrogenase solution from rabbit muscle, Type V-S, >95%, Ammonium sulfate suspension, minimum 700 units/mg protein L5132
L-Lactic Dehydrogenase from bovine muscle, Type X, Ammonium sulfate suspension, minimum 600 units/mg protein L1378
L-Lactic Dehydrogenase solution from rabbit muscle, Type XI, Salt-free, lyophilized powder, 700-1,200 units/mg protein L1254
L-Lactic Dehydrogenase isoenzymes LDH-1 (H4) and LDH-2 (H3M) from human erythrocytes, Type XII, Ammonium sulfate suspension, 300-500 units/mg protein L3379
L-Lactic Dehydrogenase solution bovine heart, Type XVII, Buffered aqueous glycerol solution, 400-600 units/mg protein L1006
L-Lactic Dehydrogenase LDH-2 (H3M) Isoenzyme from human erythrocytes, Type XX, 98% (homogeneous), Ammonium sulfate suspension, approx. 150 units/mg protein L3757
L-Lactic Dehydrogenase LDH-5 (M4) Isoenzyme solution from bovine muscle, Type XXVI, Ammonium sulfate suspension, minimum 600 units/mg protein L0508
L-Lactic Dehydrogenase LDH-5 (M4) Isoenzyme solution from porcine muscle, Type XXXII-S, Ammonium sulfate suspension, approx. 1,000 units/mg protein L5762
L-Lactic Dehydrogenase solution from rabbit muscle, Type XXXIX, Buffered aqueous glycerol solution, 700-1,000 units/mg protein L2518
L-Lactic Dehydrogenase LDH-1 (H4) Isoenzyme solution from rabbit heart, Type XXXVI, Ammonium sulfate suspension, 400-600 units/mg protein L2889
Lipase Candida rugosa, Lyophilized powder, minimum 50,000 units/mg protein, pH 7.2 (30 min incubation) L8525
Lipase human pancreas, Buffered aqueous solution, minimum 250 units/mg protein (Lowry) L9780
Lipase Mucor javanicus, Lyophilized powder, minimum 300 units/mg solid (using olive oil) L8906
Lipase from Mucor miehei, Lyophilized powder, minimum 4,000 units/mg solid (using olive oil) L9031
Lipase Pseudomonas cepacia, Lyophilized powder, minimum 50 units/g solid L9156
Lipase Rhizopus niveus, Lyophilized powder, minimum 25 units/mg solid (using olive oil) L9406
Lipase Immobilized on cellulose from Pseudomonas sp., minimum 0.5 unit/mg solid L4783
Lipase from wheat germ, Type I, Lyophilized powder, approx. 10 units/mg protein (using triacetin;, inactive on olive oil.) L3001
Lipase porcine pancreas, Type II, 100-400 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin;) L3126
Lipase Candida rugosa, Type VII, 700-1,500 units/mg solid, pH 7.2 (using olive oil (30 min incubation)) L1754
Lipase porcine pancreas, Type VI-S, Lyophilized powder, 20,000-100,000 units/mg protein (using olive oil) L0382
Lipase Rhizopus arrhizus, Type XI, Ammonium sulfate suspension, minimum 400,000 units/mg protein (using olive oil (30 min incubation)) L4384
Lipase from Chromobacterium viscosum, Type XII, Lyophilized powder, 2,000-8,000 units/mg protein (using olive oil (30 min incubation)) L0763
Lipase from Pseudomonas sp., Type XIII, Lyophilized powder, approx. 25 units/mg solid L9518
Lipase porcine pancreas, BioChemika, lyophilized, powder, ³100 U/mg 62313
Lipase porcine pancreas, BioChemika, powder, 15-35 U/mg 62300
Lipase from wheat germ, BioChemika, ~0.1 U/mg 62306
Lipase from Burkholderia sp., BioChemika, lyophilisate, slightly beige, ~12 U/mg 75577
Lipase Pseudomonas cepacia, BioChemika, powder, light beige, ~50 U/mg 62309
Lipase from Pseudomonas fluorescens, BioChemika, lyophilized, powder, grey, ~3500 U/mg 62321
Lipase from Pseudomonas fluorescens, BioChemika, EnzOS grade, powder, almost white, ~2 U/mg 71548
Lipase Pseudomonas stutzeri, BioChemika, powder, light beige, ³4 U/mg 66727
Lipase from Thermus aquaticus, BioChemika, lyophilized, powder, ~3 U/g 62293
Lipase from Thermus flavus, BioChemika, powder, brown, ~0.7 U/g 62295
Lipase Thermus thermophilus, BioChemika, powder, brown, ~0.6 U/g 62296
Lipase Candida antarctica, BioChemika, powder, light brown, ~3 U/mg 62299
Lipase Candida antarctica, BioChemika, lyophilized, powder, beige, ~0.5 U/mg 02569
Lipase A Candida antarctica, recombinant Aspergillus oryzae, BioChemika, powder, beige, ~2 U/mg 62287
Lipase B Candida antarctica, recombinant Aspergillus oryzae, BioChemika, powder, beige, ~9 U/mg 62288
Lipase Candida cylindracea, BioChemika, lyophilized, powder (fine), 15-25 U/mg 62302
Lipase Candida cylindracea, BioChemika, powder, yellow-brown, ~2 U/mg 62316
Lipase Candida lipolytica, BioChemika, powder (fine), ~1 U/g 62303
Lipase Candida rugosa, BioChemika, powder, slightly beige, >2 U/mg 90860
Lipase Candida utilis, BioChemika, powder, brown, ~0.1 U/mg 62307
Lipase from Aspergillus niger, BioChemika, powder (fine), ~200 U/g 62301
Lipase Aspergillus oryzae, BioChemika, lyophilized, powder, white, ~50 U/mg 62285
Lipase Aspergillus oryzae, BioChemika, EnzOS grade, powder, almost white, ~2 U/mg 95184
Lipase Mucor javanicus, BioChemika, powder (fine), ~10 U/mg 62304
Lipase from Mucor miehei, BioChemika, powder, slightly brown, ~1 U/mg 62298
Lipase Mucor miehei, recombinant Aspergillus oryzae, BioChemika, powder, slightly brown, ~10 U/mg 73416
Lipase Mucor miehei, recombinant Aspergillus oryzae, BioChemika, lyophilized, powder, brown, ³200 U/mg 62289
Lipase Rhizomucor miehei, recombinant Aspergillus oryzae, BioChemika, powder, beige-brown, ~0.5 U/mg 62291
Lipase Rhizopus arrhizus, BioChemika, powder (fine), ~10 U/mg 62305
Lipase Rhizopus delemar, BioChemika, ³0.4 U/mg 62328
Lipase Rhizopus niveus, BioChemika, powder (fine), ~1.5 U/mg 62310
Lipase Rhizopus oryzae, BioChemika, powder, light brown, ³30 U/mg 80612
Lipase from Penicillium camemberti, BioChemika, powder, slightly beige, ³5 U/g 96888
Lipase from Penicillium roqueforti, BioChemika, powder (fine), >0.4 U/mg 62308
Lipoprotein Lipase from bovine milk, Ammonium sulfate suspension, 2,000-6,000 units/mg protein (BCA) L2254
Luciferase from Vibrio fischeri (Photobacterium f), Lyophilized powder L8507
Luciferase from Vibrio fischeri (Photobacterium f), BioChemika, lyophilized, ~8% as protein, powder 62641
Luciferase Photinus pyralis (firefly), Lyophilized powder, 10-25 × 106 light units/mg protein L9009
Luciferase Photinus pyralis (firefly), Lyophilized powder L9506
Luciferase Photinus pyralis (firefly), Lyophilized powder, minimum 10 × 106 light units/mg protein L1759
Luciferase-Luciferin reagent in glycine buffer firefly, BioChemika, for ATP-analysis, solid, white 62646
Luciferase Photinus pyralis (firefly), BioChemika, lyophilized, ³85% as protein, powder, white 62647
Luciferase Photinus pyralis (firefly), BioChemika, soluble, ~15% as protein, powder, white 62648
Myeloperoxidase from human leukocytes, Lyophilized powder, minimum 50 units/mg protein M6908
Myeloperoxidase from human leukocytes, BioChemika, lyophilized, powder, white, ³50 U/mg protein 70021
NADH Peroxidase from Streptococcus faecalis ATCC 11700, recombinant, expressed in Escherichia coli, >95% (SDS-PAGE), Buffered aqueous solution, 50-200 units/mg protein (Biuret) N0895
Nitrate reductase (cytochrome) from Escherichia coli, Lyophilized powder, minimum 5 units/g solid N0519
Nitrate Reductase from corn seedling, minimum 4 units/mL, Buffered aqueous glycerol solution N2397
Penicillinase from Escherichia coli 205 TEM R+ (566), Lyophilized powder, 100-300 units/mg solid (using benzylpenicillin), 20-40 units/mg solid (using cephaloridine.) P3553
Penicillinase from Bacillus cereus, Type I, Lyophilized powder, 1,500-3,000 units/mg protein (using benzylpenicillin), 10-30 units/mg protein (using cephaloridine.) P0389
Penicillinase from Bacillus cereus, Type II, Lyophilized powder P6018
b-Lactamase from Enterobacter cloacae, Type III, Lyophilized powder, 50-150 units/mg protein (using cephaloridine.), 6-18 units/mg protein (using benzylpenicillin) P4399
b-Lactamase from Enterobacter cloacae, Type IV, Lyophilized powder, 0.2-0.6 units/mg protein (using benzylpenicillin) P4524
Peroxidase from Arthromyces ramosus, Lyophilized powder, 50-200 units/mg solid P4794
Peroxidase from Arthromyces ramosus, BioChemika, powder, light brown, >400 U/mg 41366
Peroxidase horseradish, Type I, Essentially salt-free, lyophilized powder, 50-150 units/mg solid P8125
Peroxidase horseradish, Type II, Essentially salt-free, lyophilized powder, 150-250 units/mg solid P8250
Peroxidase horseradish, Type VI, Essentially salt-free, lyophilized powder, 250-330 units/mg solid P8375
Peroxidase horseradish, Essentially salt-free, lyophilized powder, 250-330 units/mg solid (using pyrogallol), Type VI-A P6782
Peroxidase Highly stabilized from horseradish, Essentially salt-free, lyophilized powder, 200-300 units/mg solid P2088
Peroxidase Acidic Isoenzyme from horseradish, Type X, Ammonium sulfate suspension, approx. 250 units/mg protein (Biuret) P6140
Peroxidase horseradish, Type XII, Essentially salt-free, lyophilized powder, 250-330 units/mg solid P8415
Peroxidase from horse radish, BioChemika, lyophilized, powder, red-brown, ~850 U/mg 77334
Peroxidase from horse radish, BioChemika, lyophilized, powder, red-brown, ~700 U/mg 77333
Peroxidase from horse radish, BioChemika, powder, red-brown, ~550 U/mg 77335
Peroxidase from horse radish, BioChemika, lyophilized, powder, beige, ~150 U/mg 77332
Peroxidase from horse radish, BioChemika, essentially salt free, lyophilized, powder, light brown, ~100 U/mg 77330
Phosphatase, Acid wheat germ, minimum 0.4 unit/mg solid P3627
Phosphatase, Acid wheat germ, BioChemika, essentially salt free, lyophilized, powder, 0.1-0.6 U/mg 79410
Phosphatase, Acid, Prostatic from human semen, Lyophilized powder, minimum 200 units/mg protein P1649
Phosphatase, Acid potato, Lyophilized powder, 0.5-1.0 unit/mg solid P3752
Phosphatase, Acid potato, Lyophilized powder, 3-10 units/mg solid P1146
Phosphatase, Acid potato, Ammonium sulfate suspension, minimum 200 units/mg protein (Biuret) P0157
Phosphatase, Acid from sweet potato, Ammonium sulfate suspension, minimum 10 units/mg protein (modified Warburg-Christian) P1435
Phosphatase, Acid, Prostatic bovine semen, Lyophilized powder, approx. 2 units/g solid P3147
Phosphatase, Acid, Prostatic from bovine prostate, Lyophilized powder, approx. 10 units/g solid P6409
Phosphatase, Acid, Prostatic from human semen, Lyophilized powder, approx. 40 units/mg protein P1774
Phospho(enol)pyruvate Carboxylase corn, Ammonium sulfate suspension, minimum 1 units/mg protein P2023
Phospho(enol)pyruvate Carboxylase corn, BioChemika, powder, white, ~1 U/mg 79414
Pyruvate Kinase/Lactic Dehydrogenase enzymes from rabbit muscle, approx. 1,000 units/mL lactic dehydrogenase, approx. 700 units/mL pyruvate kinase, Buffered aqueous glycerol solution P0294
Polymyxin Acylase from Pseudomonas sp., minimum 0.1 units/mg protein (Bradford) P9208
Protease Bacillus globigii (Bacillus licheniformis), Lyophilized powder, For use in Total Dietary Fiber Assay, TDF-100A P3910
Protocatechuate 3,4-Dioxygenase from Pseudomonas sp., Lyophilized powder, 3-5 units/mg solid P8279
Pyruvate Kinase preparation from rabbit muscle, Type I, Ammonium sulfate suspension, 40-100 units/mg protein P1381
Pyruvate Kinase preparation from rabbit muscle, Type II, Ammonium sulfate suspension, 350-600 units/mg protein P1506
Pyruvate Kinase preparation from canine muscle, Type IV, Ammonium sulfate suspension, 350-500 units/mg protein P7889
Pyruvate Kinase preparation from rabbit muscle, Type VII, Buffered aqueous glycerol solution, 350-600 units/mg protein P7768
Pyruvate Kinase from Bacillus stearothermophilus, Type VIII, Lyophilized powder, 100-300 units/mg protein (Biuret) P1903
Pyruvate Kinase preparation from chicken muscle, Type X, Ammonium sulfate suspension, 300-800 units/mg protein P6406
Pyruvate Kinase preparation from rabbit muscle, BioChemika, crystalline suspension in 3.8 M ammonium sulfate solution, ~350 U/mg protein (10-40 mg/ml) 83330
Pyruvate Kinase preparation from rabbit muscle, BioChemika, lyophilized, powder, white, 100-250 U/mg 83328
Sulfatase from Helix pomatia, Type H-1, minimum 10,000 units/g solid S9626
Sulfatase solution from Helix pomatia, Type H-2, minimum 2,000 units/mL, Crude solution S9751
Sulfatase from Helix pomatia, Type H-5, Lyophilized powder S3009
Sulfatase from Patella vulgata (keyhole limpet), Type IV, Essentially salt-free, lyophilized powder, ³10 units/mg solid S8504
Sulfatase from Patella vulgata (keyhole limpet), Type V, Essentially salt-free, lyophilized powder, minimum 5 units/mg solid S8629
Sulfatase from Aerobacter aerogenes, Type VI, 10-20 units/mL, Buffered aqueous glycerol solution, 2-5 units/mg protein (Biuret) S1629
Sulfatase from abalone entrails, Type VIII, Lyophilized powder, 20-40 units/mg solid S9754
Superoxide Dismutase bovine erythrocytes, Lyophilized powder, 2,500-7,000 units/mg protein S2515
Superoxide Dismutase bovine liver, Ammonium sulfate suspension, approx. 3,000 units/mg protein (Biuret) S7008
Superoxide Dismutase bovine liver, Lyophilized powder, 2,000-6,000 units/mg protein S8160
Superoxide Dismutase canine erythrocytes, Lyophilized powder, 2,000-6,000 units/mg protein S9511
Superoxide Dismutase from Escherichia coli, Lyophilized powder, 2,500-5,000 units/mg protein S5639
Superoxide Dismutase from horseradish, Lyophilized powder, 1,000-4,000 units/mg protein