Order Center
Biochemicals
Antibiotic Explorer
BioUltra
Biomaterials
Detection
Detergent Theory
Enzyme Explorer
BioUltra Proteins
Product Indexes
Cell Signaling Enzymes
Analytical Enzymes
Enzyme Reagents
Key Resources
Lipids
Microparticles
Pharmacopoeia
PolyAmino Acids
Reference Chart
BioFiles
 Organic Synthesis & Immobilized Enzymes
Enzyme Explorer
  Immobilized Enzymes
Organic Synthesis
Product Name Product # 
Immobilized Enzymes  
a-Chymotrypsin Insoluble enzyme attached to carboxymethyl cellulose from bovine pancreas, 400-600 units/g solid C7260
Alcohol dehydrogenase–Agarose from Baker's yeast (S. cerevisiae), Lyophilized powder, 900-1,200 units/g agarose, pH 8.8, 30°C A2529
Carboxypeptidase A-Agarose, Ammonium sulfate suspension, 200-400 units/g agarose, 30°C (One ml gel will yield 6-12 units), carboxypeptidase A from bovine pancreas C1261
Catalase-Agarose, pH 5.4, Ammonium sulfate suspension, catalase from bovine liver C9284
Acetylcholinesterase-Agarose from Electrophorus electricus (electric eel), Ammonium sulfate suspension, 7,500-15,000 units/g agarose (One ml gel will yield 240-480 units) C2511
Ficin Agarose from fig tree latex, Lyophilized powder, 500-1,000 units/g agarose F1634
Lipase–Macroporous acrylic beads from Candida rugosa, Type VII-A, 300-1200 units/g solid, pH 7.2, 37°C (using olive oil) L1150
Lysozyme Agarose from chicken egg white, Lyophilized powder, 5,000-10,000 units/g solid L1129
Papain carboxymethyl cellulose from papaya latex, 150-250 units/g solid, pH 7.0, 30°C P8011
Phosphatase, Alkaline-Agarose from calf intestine, Ammonium sulfate suspension P0762
Phosphatase, Alkaline-acrylic beads from calf intestine, Ammonium sulfate suspension P0927
Proteinase K-Acrylic Beads from Tritirachium album, 2-8 units/g solid P0803
Proteinase K-Agarose from Tritirachium album, Lyophilized powder P9290
Acylase I, immobilized on Eupergit C from Aspergillus, BioChemika, ³50 U/g moist material 01824
Acylase I, immobilized on Eupergit C from Aspergillus, BioChemika, >15 U/g moist material 50837
Catalase, immobilized on Eupergit C bovine liver, BioChemika, granules, beige, >24 U/g 60639
D-Hydantoinase, recombinant, immobilized from E. coli, BioChemika, in 3.2 M (NH4)2SO4, granulated material (moist), brown-green, ~50 U/g moist material 53765
Esterase, immobilized on Eupergit® C hog liver, BioChemika, ~500 U/g moist material 46064
Lipase, immobilized in Sol-Gel-AK hog pancreas, BioChemika, ³40 U/g 62324
Lipase, immobilized in Sol-Gel-AK from Aspergillus niger, BioChemika, ³1 U/g 62281
Lipase immobilized Candida antarctica, BioChemika, beads, slightly brown, >2 U/mg 73940
Lipase, immobilized in Sol-Gel-AK on sintered glass Candida antarctica, BioChemika, beads, slightly grey, ³1 U/g 62272
Lipase, immobilized in Sol-Gel-AK Candida antarctica, BioChemika, ³1 U/g 62277
Lipase, immobilized in Sol-Gel-AK Candida cylindracea, BioChemika, ³10 U/g 62278
Lipase, immobilized in Sol-Gel-AK on sintered glass Mucor miehei, BioChemika, ³80 U/g 62274
Lipase, immobilized in Sol-Gel-AK Mucor miehei, BioChemika, ³2 U/g 62282
Lipozyme®, immobilized Mucor miehei, BioChemika, granulated material, brown, >30 U/g 62350
Lipase, immobilized in Sol-Gel-AK from Pseudomonas cepacia, BioChemika, ³40 U/g 62279
Lipase, immobilized in Sol-Gel-AK on sintered glass from Pseudomonas cepacia, BioChemika, ³10 U/g 62334
Lipase, immobilized on Ceramic particles from Pseudomonas cepacia, BioChemika, powder, light beige, ~15000 U/g 17261
Lipase, immobilized on Eupergit C from Pseudomonas fluorescens, BioChemika, granules, brown, ³0.4 U/mg 62319
Lipase, immobilized in Sol-Gel-AK on sintered glass from Pseudomonas fluorescens, BioChemika, ³10 U/g 62275
Penicillin Amidase, immobilized on Eupergit® C from E. coli, BioChemika, powder, beige, ~100 U/g 76429
Penicillin G Amidase, immobilized from E. coli, BioChemika, powder, white, ~150 U/g wet material 76428
Phosphatase alkaline, immobilized on Agarose calf intestinal mucosa, BioChemika, 600-1600 U/ml packed gel 17388
Phosphatase alkaline, immobilized on Agarose calf intestinal mucosa, BioChemika, suspension, white, ~3.5 U/mg agarose 79394
Proteinase K, immobilized on Eupergit C from Tritirachium album, BioChemika, powder (granular), ~1500 U/g 82452
Urease, immobilized on Eupergit C from jack beans, BioChemika, granules, white, ~300 U/g 94287
Organic Synthesis back to top
Acylase I porcine kidney, Grade I, Lyophilized powder, minimum 2,000 units/mg protein A3010
Aldolase from rabbit muscle, Ammonium sulfate suspension, 10-20 units/mg protein A8811
Aldolase from rabbit muscle, Lyophilized powder, approx. 10 units/mg protein A2714
Aldolase from Staphylococcus aureus, Type XII, Lyophilized powder, approx. 20 units/mg protein A2548
(+)-Camphor Monooxygenase from Pseudomonas putida, BioChemika, ~0.3 U/g 21332
Chitosanase from Streptomyces griseus, Lyophilized powder, >50 units/mg protein (Bradford) C9830
Cyclopentanone Monooxygenase from Pseudomonas sp., BioChemika, powder, off-white, ~10 U/g 29800
Cyclohexanone Monooxygenase Acinetobacter sp., BioChemika, powder, off-white, ~5 U/g 29170
Cyclohexanone Monooxygenase Nocardia globerula, BioChemika, powder, grey, ~4 U/g 29172
Cyclohexanone Monooxygenase Xanthobacter sp., BioChemika, powder, off-white, ~6 U/g 29174
2-Deoxyribose-5-phosphate Aldolase from Lactobacillus plantarum, BioChemika, powder, slightly yellow, ~15 U/g 41228
Epoxide Hydrolase Rhodococcus rhodochrous, BioChemika, lyophilisate, beige, ³0.5 U/g 45299
Epoxide Hydrolase, Aspergillus niger sp., recombinant E. coli, BioChemika, powder, light-brown, ~35 U/g 71832
Esterase solution porcine liver, Ammonium sulfate suspension, minimum 150 units/mg protein (Biuret) E2884
Esterase from porcine liver, Lyophilized powder, minimum 15 units/mg solid E3019
Esterase rabbit liver, Ammonium sulfate suspension, 80-120 units/mg protein (Biuret) E9636
Esterase rabbit liver, Lyophilized powder, approx. 100 units/mg protein E0887
Esterase from porcine liver, BioChemika, lyophilized, powder, slightly beige, ³130 U/mg 46058
Esterase from porcine liver, BioChemika, suspension in 3.2 M ammonium sulfate solution, ~130 U/mg protein (~10 mg/ml) 46063
Esterase Isoenzyme 1 hog liver, BioChemika, suspension in 3.2 M ammonium sulfate solution, ~200 U/mg protein (~15 mg/ml) 46048
Esterase from horse liver, BioChemika, lyophilized, powder, brown, 0.5-1.0 U/mg 46069
Esterase from Bacillus sp., BioChemika, ~0.1 U/mg 46062
Esterase from Bacillus stearothermophilus, BioChemika, ~0.4 U/mg 46051
Esterase Bacillus thermoglucosidasius, BioChemika, lyophilized, powder, beige, ~0.1 U/mg 46054
Esterase Streptomyces diastatochromogenes, recombinant from E. coli, BioChemika, powder, slightly beige, ³50 U/mg 78042
Esterase from Thermoanaerobium brockii, BioChemika, powder, off-white, ~2 U/g 46061
Esterase Candida lipolytica, BioChemika, ~0.1 U/mg 46056
Esterase Saccharomyces cerevisiae, BioChemika, lyophilized, powder, beige, ~2 U/g 46071
Esterase Mucor miehei, BioChemika, powder, slightly brown, ~1 U/mg 46059
Esterase Rhizomucor miehei, recombinant Aspergillus oryzae, BioChemika, solution, very deep brown, ³250 U/ml 75900
Esterase Rhizopus oryzae, BioChemika, powder, light-beige, ³20 U/g 79208
Esterase Thermomyces lanuginosus, recombinant Aspergillus oryzae, BioChemika, solution, light brown, >1200 U/ml 27637
Aldolase from rabbit muscle, BioChemika, powder, white, 15-20 U/mg 05518
Aldolase from rabbit muscle, BioChemika, crystalline suspension in 3.2 M ammonium sulfate solution, pH ~6, ~9 U/mg protein (~10 mg/ml) 05520
Aldolase from Staphylococcus aureus, BioChemika, powder, white, ³10 U/mg 05522
Aldolase from Thermus aquaticus, BioChemika, lyophilisate, yellow, ~5 U/g 05525
Fructose-1,6-bisphosphate Aldolase from Bacillus subtilis, BioChemika, lyophilisate, slightly beige, ~30 U/g 80753
Fructose-1,6-bisphosphate Aldolase Staphylococcus carnosus, BioChemika, lyophilisate, slightly beige, ~0.4 U/mg 94864
a-Galactosidase, positionally specific solution from Escherichia coli, recombinant, expressed in Escherichia coli, Buffered aqueous solution G7163
b-1®4-Galactosidase, positionally specific, recombinant, expressed in Escherichia coli, Buffered aqueous solution G0413
b-1®6-Galactosidase, positionally specific, recombinant, expressed in Escherichia coli, Buffered aqueous solution G0914
b-1®(3,4,6)-Galactosidase, positionally specific, recombinant, expressed in Escherichia coli, Buffered aqueous solution G1288
2-Hydroxybiphenyl 3-Monooxygenase recombinant, BioChemika, solution, slightly yellow, ~2 U/ml 93043
4-Hydroxy-2-oxoglutarate Aldolase from E. coli, BioChemika, lyophilisate, white, ~0.1 U/mg 59892
Lipase Candida rugosa, Lyophilized powder, minimum 50,000 units/mg protein, pH 7.2 (30 min incubation) L8525
Lipase human pancreas, Buffered aqueous solution, minimum 250 units/mg protein (Lowry) L9780
Lipase Mucor javanicus, Lyophilized powder, minimum 300 units/mg solid (using olive oil) L8906
Lipase from Mucor miehei, Lyophilized powder, minimum 4,000 units/mg solid (using olive oil) L9031
Lipase Pseudomonas cepacia, Lyophilized powder, minimum 50 units/g solid L9156
Lipase Rhizopus niveus, Lyophilized powder, minimum 25 units/mg solid (using olive oil) L9406
Lipase Immobilized on cellulose from Pseudomonas sp., minimum 0.5 unit/mg solid L4783
Lipase from wheat germ, Type I, Lyophilized powder, approx. 10 units/mg protein (using triacetin;, inactive on olive oil.) L3001
Lipase porcine pancreas, Type II, 100-400 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin;) L3126
Lipase Candida rugosa, Type VII, 700-1,500 units/mg solid, pH 7.2 (using olive oil (30 min incubation)) L1754
Lipase porcine pancreas, Type VI-S, Lyophilized powder, 20,000-100,000 units/mg protein (using olive oil) L0382
Lipase Rhizopus arrhizus, Type XI, Ammonium sulfate suspension, minimum 400,000 units/mg protein (using olive oil (30 min incubation)) L4384
Lipase from Chromobacterium viscosum, Type XII, Lyophilized powder, 2,000-8,000 units/mg protein (using olive oil (30 min incubation)) L0763
Lipase from Pseudomonas sp., Type XIII, Lyophilized powder, approx. 25 units/mg solid L9518
Lipase porcine pancreas, BioChemika, lyophilized, powder, ³100 U/mg 62313
Lipase porcine pancreas, BioChemika, powder, 15-35 U/mg 62300
Lipase from wheat germ, BioChemika, ~0.1 U/mg 62306
Lipase from Burkholderia sp., BioChemika, lyophilisate, slightly beige, ~12 U/mg 75577
Lipase Pseudomonas cepacia, BioChemika, powder, light beige, ~50 U/mg 62309
Lipase from Pseudomonas fluorescens, BioChemika, lyophilized, powder, grey, ~3500 U/mg 62321
Lipase from Pseudomonas fluorescens, BioChemika, EnzOS grade, powder, almost white, ~2 U/mg 71548
Lipase Pseudomonas stutzeri, BioChemika, powder, light beige, ³4 U/mg 66727
Lipase from Thermus aquaticus, BioChemika, lyophilized, powder, ~3 U/g 62293
Lipase from Thermus flavus, BioChemika, powder, brown, ~0.7 U/g 62295
Lipase Thermus thermophilus, BioChemika, powder, brown, ~0.6 U/g 62296
Lipase Candida antarctica, BioChemika, powder, light brown, ~3 U/mg 62299
Lipase Candida antarctica, BioChemika, lyophilized, powder, beige, ~0.5 U/mg 02569
Lipase A Candida antarctica, recombinant Aspergillus oryzae, BioChemika, powder, beige, ~2 U/mg 62287
Lipase B Candida antarctica, recombinant Aspergillus oryzae, BioChemika, powder, beige, ~9 U/mg 62288
Lipase Candida cylindracea, BioChemika, lyophilized, powder (fine), 15-25 U/mg 62302
Lipase Candida cylindracea, BioChemika, powder, yellow-brown, ~2 U/mg 62316
Lipase Candida lipolytica, BioChemika, powder (fine), ~1 U/g 62303
Lipase Candida rugosa, BioChemika, powder, slightly beige, >2 U/mg 90860
Lipase Candida utilis, BioChemika, powder, brown, ~0.1 U/mg 62307
Lipase from Aspergillus niger, BioChemika, powder (fine), ~200 U/g 62301
Lipase Aspergillus oryzae, BioChemika, lyophilized, powder, white, ~50 U/mg 62285
Lipase Aspergillus oryzae, BioChemika, EnzOS grade, powder, almost white, ~2 U/mg 95184
Lipase Mucor javanicus, BioChemika, powder (fine), ~10 U/mg 62304
Lipase from Mucor miehei, BioChemika, powder, slightly brown, ~1 U/mg 62298
Lipase Mucor miehei, recombinant Aspergillus oryzae, BioChemika, powder, slightly brown, ~10 U/mg 73416
Lipase Mucor miehei, recombinant Aspergillus oryzae, BioChemika, lyophilized, powder, brown, ³200 U/mg 62289
Lipase Rhizomucor miehei, recombinant Aspergillus oryzae, BioChemika, powder, beige-brown, ~0.5 U/mg 62291
Lipase Rhizopus arrhizus, BioChemika, powder (fine), ~10 U/mg 62305
Lipase Rhizopus delemar, BioChemika, ³0.4 U/mg 62328
Lipase Rhizopus niveus, BioChemika, powder (fine), ~1.5 U/mg 62310
Lipase Rhizopus oryzae, BioChemika, powder, light brown, ³30 U/mg 80612
Lipase from Penicillium camemberti, BioChemika, powder, slightly beige, ³5 U/g 96888
Lipase from Penicillium roqueforti, BioChemika, powder (fine), >0.4 U/mg 62308
Lipoprotein Lipase Chromobacterium viscosum, BioChemika, lyophilized, powder, ~2500 U/mg (after reconstitution) 62333
Lipoprotein Lipase from Pseudomonas sp., BioChemika, lyophilized, powder, yellow-brown, ~160 U/mg 62336
Lipoprotein Lipase from Pseudomonas sp., BioChemika, lyophilized, powder, 1500-2500 U/mg 62335
(S)-Oxynitrilase from Manihot esculenta (Maniok), BioChemika, solution (turbid), ³3000 U/ml 71811
Phospholipase A2 honey bee venom, Salt-free, lyophilized powder P9279
Phospholipase A2 bovine pancreas, Lyophilized powder P8913
Phospholipase A2 Crotalus atrox venom, Lyophilized powder, 300-700 units/mg protein, pH 8.9, 25°C (using soybean L-a-phosphatidylcholine) P3770
Phospholipase A2 from Streptomyces violaceoruber, Lyophilized powder, approx. 200 units/mg protein, pH 8.9, 25°C (using soybean L-a-phosphatidylcholine) P5910
Phospholipase A2 Naja mossambica mossambica, Lyophilized powder, approx. 1,500 units/mg protein, pH 8.9, 25°C (using soybean L-a-phosphatidylcholine) P4034
Phospholipase A2 Naja mossambica mossambica, Lyophilized powder, minimum 300 units/mg protein, pH 8.9, 25°C (using soybean L-a-phosphatidylcholine) P7653
Phospholipase A2 Naja mossambica mossambica, Lyophilized powder, approx. 1,500 units/mg protein, pH 8.9, 25°C (using soybean L-a-phosphatidylcholine) P7778
Phospholipase A2 Naja naja venom, Lyophilized powder P6139
Phospholipase A2 from porcine pancreas, Ammonium sulfate suspension, minimum 600 units/mg protein, pH 8.0, 37°C (using soybean L-a-phosphatidylcholine) P6534
Phospholipase A2 Streptomyces violaceoruber, Lyophilized powder, minimum 10 units/mg solid, pH 8.0, 37°C P8685
Phospholipase A2 Agkistrodon halys, BioChemika, powder, white, ~11 U/mg 79475
Phospholipase A2 bovine pancreas, BioChemika, 20%, powder, off-white, ~7 U/mg 79476
Phospholipase A2 honey bee venom, BioChemika, powder, off-white, ³1000 U/mg 79478
Phospholipase A2 from porcine pancreas, BioChemika, suspension, white, >600 U/mg protein (~3 mg/ml) 79482
Phospholipase A2 from porcine pancreas, BioChemika, lyophilized, powder, white, ~500 U/mg 79483
Phospholipase B from Vibrio sp., Lyophilized powder, minimum 20 units/mg protein P8914
Phospholipase C from Bacillus cereus, Type III, Lyophilized powder, 100-200 units/mg protein (using soybean L-a-phosphatidylcholine) P6135
Phospholipase C from Clostridium perfringens (C. welchii), Type I-S, Lyophilized powder P9185
Phospholipase C from Bacillus cereus, Lyophilized powder, 500-2,000 units/mg protein (using egg yolk phosphatidylcholine) P9439
Phospholipase C from Clostridium perfringens (C. welchii), Type IX, Lyophilized powder P1392
Phospholipase C from Bacillus cereus, Type XI, Ammonium sulfate suspension P7147
Phospholipase C from Clostridium perfringens (C. welchii), Type XIV, Lyophilized powder P4039
Phospholipase C, Phosphatidylinositol-specific from Bacillus cereus, Buffered aqueous glycerol solution, 3,000 units/mg protein, minimum 3,000 units/mg protein (Lowry) P8804
Phospholipase C from Bacillus cereus, BioChemika, powder, yellow-brown, ~12 U/mg 79477
Phospholipase C from Clostridium perfringens (C. welchii), BioChemika, powder, slightly beige, 30-200 U/mg 79485
Phospholipase C from Clostridium perfringens (C. welchii), BioChemika, lyophilized, powder, brown, ~4 U/mg 79487
Phospholipase D from Arachis hypogaea (peanut), Type II, Lyophilized powder, minimum 60 units/mg protein P0515
Phospholipase D from Streptomyces chromofuscus, Lyophilized powder, minimum 2,000 units/mg solid P8023
Phospholipase D from Streptomyces sp., Type VII, Lyophilized powder, minimum 150 units/mg solid P4912
Phospholipase D from cabbage, Type IV, Lyophilized powder, minimum 200 units/mg solid P8398
Progesterone Monooxygenase from Cylindrocapron radicicola, BioChemika, powder, beige, ~0.6 U/g 81703
Superoxide Dismutase bovine erythrocytes, BioChemika, powder, blue-grey, 2'000-10'000 U/mg 86200
L-Threonine Aldolase Candida humicola, BioChemika, Lyophilisate, slightly beige, ~10 U/g 53806
L-Threonine Aldolase from Pseudomonas putida, BioChemika, Lyophilisate, slightly yellow, ~0.2 U/mg 96586
Transaminase, broad-range bacterial, recombinant, expressed in Escherichia coli, Lyophilized powder, minimum 5 units/mg solid T7684
2-Tridecanone Monooxygenase from Pseudomonas cepacia, BioChemika, ~0.5 U/g 91530
Tyrosinase mushroom, ³2,000 units/mg solid T7755
b-Tyrosinase, BioChemika, solution, deep yellow, ³25 U/mg protein (~6 mg/ml) 40522
Tyrosinase mushroom, BioChemika, lyophilized, powder, brown, ³2000 U/mg 93898