Order Center
Biochemicals
Antibiotic Explorer
BioUltra
Biological Detergents
Detection
Enzyme Explorer
BioUltra Proteins
Product Indexes
Cell Signaling Enzymes
Analytical Enzymes
Enzyme Reagents
Key Resources
Lipids
Microparticles
Peptide Explorer
Pharmacopoeia
PolyAmino Acids
BioFiles
 Proteases & Protein Sequencing
Enzyme Explorer
  Fusion Protein Cleavage
Proteases
Protein Sequencing
Product Name Product # 
Fusion Protein Cleavage  
Elastase, pancreatic from porcine pancreas, Type IV, Lyophilized powder, minimum 4 units/mg protein (Biuret) E0258
Elastase from human leukocytes, Lyophilized powder, ³50 units/mg protein (Bradford) E8140
Elastase, pancreatic solution from porcine pancreas, Type I, Aqueous suspension, minimum 4 units/mg protein E1250
Elastase, pancreatic from porcine pancreas, Type II-A, Lyophilized powder, minimum 1 units/mg protein (Biuret) E6883
Elastase, pancreatic from porcine pancreas, Type III, Lyophilized powder, minimum 4 units/mg protein E0127
Elastase, pancreatic from porcine pancreas, BioChemika, powder, white, ³15 U/mg 45124
Elastase, pancreatic from porcine pancreas, BioChemika, lyophilized, powder, white, ~8 U/mg 45125
Elastase, pancreatic from porcine pancreas, BioChemika, powder, off-white, ³3 U/mg 45122
Elastase, pancreatic from porcine pancreas, BioChemika, crystalline suspension with 0.01% thymol as preservative, 1-2 U/mg protein (~2 mg/ml) 45127
Elastase, pancreatic from porcine pancreas, Type IV, Lyophilized powder, minimum 4 units/mg protein (Biuret) E0258
Enterokinase from bovine intestine, Powder E5144
Enterokinase from bovine intestine, Salt-free, lyophilized powder, ³0.5 units/mg solid E5510
Enterokinase solution from calf intestine, Buffered aqueous glycerol solution, minimum 1,000 units/mg protein (BCA) E0766
Enterokinase porcine intestine, Salt-free, lyophilized powder, approx. 2 units/mg solid E0632
Enterokinase porcine intestine, Lyophilized powder, 100-500 units/mg protein E0885
Factor X from bovine plasma, Buffered aqueous solution, approx. 80 units/mg protein (E1%/280 = 12.4) F4003
Thrombin bovine plasma, Lyophilized powder, 175-350 NIH units/mg protein (Biuret) T4265
Thrombin bovine plasma, Lyophilized powder, 50-175 NIH units/mg protein (Biuret) T4648
Thrombin bovine plasma, Lyophilized powder T7513
Thrombin from human plasma, Lyophilized powder T1063
Thrombin from human plasma, Lyophilized powder, approx. 2,000 NIH units/mg protein (E1%/280 = 18.3) T6884
Thrombin from human plasma, Lyophilized powder, approx. 1,000 NIH units/mg protein (E1%/280 = 18.3) T7009
Thrombin from human plasma, suitable (For routine use in the thrombin time test), Lyophilized powder T8885
Thrombin from human plasma, suitable (For routine use in the thrombin time test.), Lyophilized powder T9135
Thrombin from human plasma, Lyophilized powder, 50-300 NIH units/mg protein T9677
Thrombin from human plasma, BioChemika, lyophilized, powder, white, ~0.8 U/mg 89223
Thrombin bovine plasma, BioChemika, powder, white, ~1 U/mg 89228
Proteases back to top
Achromopeptidase from Achromobacter lyticus, Crude powder, 800-3,200 units/mg solid A3547
Achromopeptidase from Achromobacter lyticus, Crude powder, 300-600 units/mg solid A7550
Achromopeptidase from Achromobacter lyticus, Partially purified powder, 20,000-40,000 units/mg solid A3422
a-Chymotrypsin DFP treated, Type VI, Lyophilized powder, <0.2 units/mg protein C1012
Agkistrodon contortrix thrombin-like enzyme, Lyophilized powder A8766
Aminopeptidase I from Streptomyces griseus, Lyophilized powder, minimum 200 units/mg protein A9934
Ancrod from Agkistrodon rhodostoma (Malayan pit viper) venom, Lyophilized powder, minimum 200 NIH units/mg protein (E1%/280 = 14.8) A5042
Bromelain pineapple stem, Lyophilized powder, 5-10 units/mg protein B5144
Bromelain pineapple stem, BioChemika, powder, yellow-brown, 2-5 U/mg 16990
Bromoperoxidase Corallina officinalis, Lyophilized powder, minimum 100 units/mg protein (Lowry) B2170
Carboxypeptidase A-Agarose, Ammonium sulfate suspension, 200-400 units/g agarose, 30°C (One ml gel will yield 6-12 units), carboxypeptidase A from bovine pancreas C1261
Carboxypeptidase A bovine pancreas, Type I, Aqueous suspension, approx. 50 units/mg protein C0261
Carboxypeptidase A bovine pancreas, bovine pancreas, Aqueous suspension, approx. 50 units/mg protein C0386
Carboxypeptidase A, PMSF treated bovine pancreas, bovine pancreas, approx. 50 units/mg protein C9268
Carboxypeptidase A bovine pancreas, BioChemika, suspension, white, ³1100 U/ml 21937
Carboxypeptidase A bovine pancreas, BioChemika, suspension, white, ~70 U/mg protein (~20 mg/ml) 21938
Carboxypeptidase A, PMSF treated bovine pancreas, BioChemika, suspension, white, ~60 U/mg protein (~20 mg/ml) 21940
Carboxypeptidase B porcine pancreas, Lyophilized powder, 175-250 units/mg protein C9584
Carboxypeptidase G from Pseudomonas sp., Lyophilized powder, minimum 3 units/mg protein C9658
Carboxypeptidase P from Penicillium janthinellum, Lyophilized powder C5396
Carboxypeptidase P from Penicillium janthinellum, BioChemika, lyophilized, powder, white, ³18 U/mg 21946
Carboxypeptidase W from wheat, 20-100 units/mg solid C6527
Carboxypeptidase Y from Baker's yeast (S. cerevisiae), Lyophilized powder, approx. 100 units/mg protein C3888
Carboxypeptidase Y from Baker's yeast (S. cerevisiae), BioChemika, ~20% as protein, powder, light brown, ~20 U/mg 21943
Carboxypeptidase Y from Baker's yeast (S. cerevisiae), BioChemika, lyophilized, ~10% as protein, powder, ~12 U/mg 21945
Caspase 3 human, minimum 90% (SDS-PAGE), recombinant, expressed in Escherichia coli (C-terminal histidine-tagged), Buffered aqueous glycerol solution, >1,000 units/mg protein C1224
Cathepsin B from bovine spleen, Lyophilized powder, minimum 10 units/mg protein C6286
Cathepsin B from human liver, Buffered aqueous solution, 1,000-4,000 units/mg protein (E1%/280) C8571
Cathepsin B from human placenta, Lyophilized powder, minimum 5 units/mg protein C0150
Cathepsin C from bovine spleen, Type X, Lyophilized powder, approx. 10 units/mg protein C8511
Cathepsin D from bovine spleen, Lyophilized powder, minimum 5 units/mg protein C3138
Cathepsin D from human liver, Lyophilized powder, minimum 300 units/mg protein (E1%/280) C8696
Cathepsin G from human leukocytes, Lyophilized powder, minimum 60 units/mg protein (Bradford) C4428
Chymopapain from papaya latex, Lyophilized powder, 0.5-2.0 units/mg protein C8526
a-Chymotrypsin from human pancreas, Lyophilized powder C8946
a-Chymotrypsin Insoluble enzyme attached to carboxymethyl cellulose from bovine pancreas, 400-600 units/g solid C7260
a-Chymotrypsin-Agarose from bovine pancreas, Lyophilized powder, 2,000-3,500 units/g agarose (One ml gel will yield 65-120 units) C9134
a-Chymotrypsin from bovine pancreas, Type II, Essentially salt-free, lyophilized powder, 40-60 units/mg protein C4129
a-Chymotrypsin from bovine pancreas, Type I-S, Essentially salt-free, lyophilized powder, 40-60 units/mg protein C7762
a-Chymotrypsin TLCK Treated from bovine pancreas, Type VII, Essentially salt-free, lyophilized powder, 40-60 units/mg protein C3142
a-Chymotrypsin-polyethylene glycol, approx. 20 BTEE units/mg protein C5088
a-Chymotrypsin from bovine pancreas, BioChemika, for protein sequence analysis, lyophilisate (extensively salt-free) 27207
a-Chymotrypsin from bovine pancreas, BioChemika, ~65 U/mg 27267
Ethylenediaminetetraacetic acid disodium salt dihydrate, purum, 98-101% (complexometric) 27270
a-Chymotrypsin from bovine pancreas, BioChemika, lyophilized, powder, ~70 U/mg 27270
a-Chymotrypsin from bovine pancreas, BioChemika, powder, almost white, ~50 U/mg 27272
a-Chymotrypsin, TLCK treated, BioChemika, lyophilized, powder, white, 60 U/mg 27280
b-Chymotrypsin from bovine pancreas, Essentially salt-free, lyophilized powder, 20-50 units/mg solid C4629
a-Chymotrypsinogen A from bovine pancreas, Type II, Essentially salt-free, lyophilized powder, 40-60 units/mg solid (after activation to a-chymotrypsin.) C4879
Clostripain from Clostridium histolyticum, Lyophilized powder, 200-400 units/mg protein C0799
Clostripain from Clostridium histolyticum, 20-100 units/mg solid C0888
Clostripain from Clostridium histolyticum, Lyophilized powder, 200-400 units/mg protein C7403
Clostripain from Clostridium histolyticum, BioChemika, lyophilized, ~80% as protein, powder, white, >100 U/mg protein 27549
Collagenase from Clostridium histolyticum, 1.8-2.2 FALGPA units/mg protein (Neutral Protease: <10 units per mg solid), Sigma Blend Type F C7926
Collagenase from Clostridium histolyticum, 1.1-1.5 FALGPA units/mg protein (Neutral Protease: 20-50units per mg solid), Sigma Blend Type H C8051
Collagenase from Clostridium histolyticum, 0.5-0.9 FALGPA units/mg protein (Neutral Protease: 50-80 units per mg solid), Sigma Blend Type L C8176
Collagenase from Clostridium histolyticum, Lyophilized powder, Type VII-S C2399
Collagenase from Clostridium histolyticum, Lyophilized powder, Type XI-S C4785
Collagenase from Clostridium histolyticum, Essentially salt-free, lyophilized powder, 0.25-1.0 FALGPA units/mg protein, >125 CDU/mg solid (CDU = collagen digestion units), Type I C0130
Collagenase from Clostridium histolyticum, Essentially salt-free, lyophilized powder, 0.5-3.0 FALGPA units/mg protein, >125 CDU/mg solid (CDU = collagen digestion units), Type IA C9891
Collagenase from Clostridium histolyticum, Lyophilized powder, Type IA-S C5894
Collagenase from Clostridium histolyticum, 0.5-3.0 FALGPA units/mg solid, >125 CDU/mg solid (CDU = collagen digestion units), Type II C6885
Collagenase from Clostridium histolyticum, Type III, Fraction A, Lyophilized powder, 2-10 FALGPA units/mg protein, minimum 400 CDU/mg solid (CDU = collagen digestion units) C0255
Collagenase from Clostridium histolyticum, Lyophilized powder, Type II-S C1764
Collagenase from Clostridium histolyticum, Lyophilized powder, Type I-S C1639
Collagenase from Clostridium histolyticum, 0.5-3.0 FALGPA units/mg solid, >125 CDU/mg solid (CDU = collagen digestion units), Type IV C5138
Collagenase from Clostridium histolyticum, Lyophilized powder, Type IV-S C1889
Collagenase from Clostridium histolyticum, 1-3 FALGPA units/mg protein, >125 CDU/mg solid (CDU = collagen digestion units), Type V C9263
Collagenase from Clostridium histolyticum, Type VII, Lyophilized powder, 1,000-3,000 CDU/mg solid (CDU = collagen digestion units), 4-12 FALGPA units/mg protein C0773
Collagenase from Clostridium histolyticum, suitable for release of rat epididymal adipocytes and hepatocytes (for methodology see Type II and Type IV), 0.5-3.0 FALGPA units/mg solid, >125 CDU/mg solid (CDU = collagen digestion units), Type VIII C2139
Collagenase from Clostridium histolyticum, Lyophilized powder, Type V-S C2014
Collagenase from Clostridium histolyticum, 2-5 FALGPA units/mg solid, >1200 CDU/mg solid (CDU = collagen digestion units), Type XI C7657
Collagenase/Dispase from Achromobacter iophagus/Bacillus polymyxa C3180
Collagenase from Clostridium histolyticum, BioChemika, lyophilized, powder, 0.15-0.4 U/mg 27665
Collagenase from Clostridium histolyticum, BioChemika, powder, light brown, ~0.1 U/mg 27661
Collagenase from Clostridium histolyticum, BioChemika, powder, brown, 0.4-2 U/mg 27678
Cucumisin, Lyophilized powder, 10-30 units/mg solid C1932
Dipeptidyl Peptidase from porcine kidney, Lyophilized powder, minimum 10 units/mg protein (Bradford) D7052
Elastase, pancreatic solution from porcine pancreas, Type I, Aqueous suspension, minimum 4 units/mg protein E1250
Elastase, pancreatic from porcine pancreas, Type III, Lyophilized powder, minimum 4 units/mg protein E0127
Elastase, pancreatic from porcine pancreas, BioChemika, powder, white, ³15 U/mg 45124
Elastase, pancreatic from porcine pancreas, BioChemika, lyophilized, powder, white, ~8 U/mg 45125
Elastase, pancreatic from porcine pancreas, BioChemika, powder, off-white, ³3 U/mg 45122
Elastase, pancreatic from porcine pancreas, BioChemika, crystalline suspension with 0.01% thymol as preservative, 1-2 U/mg protein (~2 mg/ml) 45127
ELISA-Systems aflatoxine M1 45169
Endoproteinase, recombinant from Bacillus sp., BioChemika, powder, slightly beige, ³40 U/mg 45169
Endoproteinase Asp-N from Pseudomonas fragi (mutant), BioChemika 45168
Endoproteinase Glu-C Staphylococcus aureus V8, BioChemika, lyophilized, powder, light brown, 400-1000 U/mg 45172
ELISA-Systems aflatoxine B1 45172
Endoproteinase Lys-C from Lysobacter enzymogenes, BioChemika, lyophilized, powder, ~3 U/vial 45175
ELISA-Systems clean-up column II 45175
Enterokinase from bovine intestine, Salt-free, lyophilized powder, ³0.5 units/mg solid E5510
Enterokinase solution from calf intestine, Buffered aqueous glycerol solution, minimum 1,000 units/mg protein (BCA) E0766
Enterokinase porcine intestine, Salt-free, lyophilized powder, approx. 2 units/mg solid E0632
Enterokinase porcine intestine, Lyophilized powder, 100-500 units/mg protein E0885
Factor V Activating Enzyme from Russell”s viper venom, Lyophilized powder, 20 units/mg protein (E1%/280 = 10.0) F3380
Factor X from bovine plasma, Buffered aqueous solution, approx. 80 units/mg protein (E1%/280 = 12.4) F4003
Ficin from fig tree latex, Saline suspension, minimum 1.5 units/mg protein (Biuret) F4125
Ficin from fig tree latex, Lyophilized powder, minimum 1 units/mg protein F6008
Ficin from fig tree latex, Powder, minimum 0.1 unit/mg solid F4165
Ficin Agarose from fig tree latex, Lyophilized powder, 500-1,000 units/g agarose F1634
Glycopeptidase from almonds, minimum 0.05 unit/mL, Buffered aqueous glycerol solution G0535
Kallikrein from human plasma, Lyophilized powder, 1-5 units/mg protein K1004
Kallikrein from human plasma, Buffered aqueous glycerol solution, 5-15 units/mg protein (Lowry) K3126
Kallikrein from porcine pancreas, minimum 40 units/mg solid K3627
Keratanase from Pseudomonas sp., Lyophilized powder K2876
Leucine Aminopeptidase, microsomal from porcine kidney, Type VI, Lyophilized powder, approx. 25 units/mg protein (E1%/280) L0632
Leucine Aminopeptidase, microsomal from porcine kidney, Type VI-S, Lyophilized powder, 15-25 units/mg protein (Biuret) L6007
Leucine Aminopeptidase, microsomal from porcine kidney, BioChemika, powder, white, ~18 U/mg 61858
Leucine Aminopeptidase, microsomal from porcine kidney, BioChemika, lyophilized, powder, off-white, ~10 U/mg 61857
Leucine Aminopeptidase, cytosol porcine kidney, Ammonium sulfate suspension, minimum 50 units/mg protein L5658
Leucine Aminopeptidase, cytosol porcine kidney, BioChemika, crystalline suspension in 3.8 M ammonium sulfate solution, >80 U/mg protein (~5-15 mg/ml) 61860
Leucine Aminopeptidase, microsomal from porcine kidney, Type IV-S, Ammonium sulfate suspension, 10-40 units/mg protein (Bradford) L5006
Leucozyme from human sputum L8413
Matrix Metalloproteinase-1 from human fibroblasts, Solution M1802
Matrix Metalloproteinase-2 from human fibroblasts, Solution M1927
Matrix Metalloproteinase-3 from human fibroblasts, Solution M1677
Matrix Metalloproteinase-9 from mouse, recombinant, expressed in BHK cells, Buffered aqueous glycerol solution M1552
Metalloendopeptidase from Grifola frondosa M4394
Protease from Bacillus amyloliquefaciens, Liquid, minimum 0.8 U/g P1236
Papain from papaya latex P5306
Papain from papaya latex, 0.5-2 units/mg solid P3250
Papain from papaya latex, Crude powder, 1.5-3.5 units/mg solid P3375
Papain Agarose preparation from papaya latex, Lyophilized powder, 90-150 units/ml gel P4406
Papain from papaya latex, minimum 10 units/mg protein (E1%/280) P4762
Papain Mercuripapain from papaya latex, Aqueous ethanol suspension, 16-40 BAEE units/mg protein (Biuret) P9886
Papain from papaya latex, Buffered aqueous suspension, 16-40 units/mg protein P3125
Papain Carica papaya, BioChemika, solution, light brown, 10-20 U/mg protein (~25 mg/ml) 76216
Papain Carica papaya, BioChemika, powder, almost white, ~12 U/mg 76218
Papain Carica papaya, BioChemika, powder, ~3 U/mg 76220
Papain Carica papaya, BioChemika, powder, ~0.5 U/mg 76222
Pepsin porcine gastric mucosa, Powder, 800-2,500 units/mg protein P7000
Pepsin porcine gastric mucosa, Powder, 600-1,800 units/mg protein P7125
Pepsin porcine gastric mucosa, Lyophilized powder, 2,500-3,500 units/mg protein (E1%/280) P7012
Pepsin porcine gastric mucosa, Lyophilized powder, 3,200-4,500 units/mg protein P6887
Pepsin porcine gastric mucosa, BioChemika, 2×cryst., lyophilized, powder, ³2400 U/mg 77152
Pepsin porcine gastric mucosa, BioChemika, powder, slightly beige, 1200-2400 U/mg 77151
Pepsin porcine gastric mucosa, BioChemika, powder, slightly beige, 600-1200 U/mg 77160
Pepsin porcine gastric mucosa, BioChemika, powder, light-beige, 200-600 U/mg 77163
Pepsinogen porcine stomach, Grade I-S, Lyophilized powder, approx. 3,000 units/mg protein (after activation to pepsin at pH 2.0 at 25°C) P4656
Pepsinogen I human P1490
Pepsinogen II human gastric mucosa P1615
Peptidase from porcine intestinal mucosa, 50-100 units/g solid P7500
Plasmin from porcine blood, 3-5 units/mg protein (Biuret) P8644
Plasmin from human plasma, BioChemika, lyophilized, powder, white, >4 U/mg 80955
Plasminogen from human plasma, not suitable for streptokinase determination by clot formation procedure, Lyophilized powder, 6-9 units/mg protein P7397
Protease Streptomyces griseus, BioChemika, lyophilized, powder, slightly brown, ~6 U/mg 81748
Protease Streptomyces griseus, BioChemika, powder, light brown, ~5 U/mg 81750
Proteinase from Bacillus subtilis, BioChemika, powder, slightly beige, >2 U/mg 96887
Protease Bacillus globigii (Bacillus licheniformis), Lyophilized powder, For use in Total Dietary Fiber Assay, TDF-100A P3910
Protease from Aspergillus oryzae, minimum 500 U/g P6110
Protease from Bacillus globigii (Bacillus licheniformis), minimum 2.4 U/g P4860
Protease Bacillus globigii (Bacillus licheniformis), Aqueous propylene glycol solution, 7-15 units/mg protein (Biuret) P5459
Protease Bacillus sp., minimum 16 U/g P3111
Protease Bacillus sp., minimum 8 U/g P5860
EverlaseTM 16.0 L, minimum 16 U/g P5985
Endoproteinase Lys-C from Lysobacter enzymogenes, Lyophilized powder P2289
Protease from HIV-1, suitable for cleaving HIV substrate III (Yields 2 peaks which elute before the substrate peak when analyzed by reversed phase HPLC.), recombinant, expressed in Escherichia coli, Buffered aqueous solution P7338
Protease S from Pyrococcus furiosis, recombinant, expressed in Bacillus sp., approx. 9.5 units/mg protein P6361
Protease from bovine pancreas, Type I, minimum 5 units/mg solid P4630
Protease from Bacillus polymyxa, Type IX, approx. 1.0 unit/mg solid P6141
Subtilisin A, Type VIII, Lyophilized powder, 7-15 units/mg solid P5380
Protease-Agarose from Bacillus licheniformis, Type VIII-A, Lyophilized powder, 75-150 units/g agarose P8790
Thermolysin Bacillus thermoproteolyticus rokko, Type X, Lyophilized powder, 50-100 units/mg protein (E1%/280) P1512
Protease from Aspergillus saitoi, Type XIII, minimum 0.6 unit/mg solid P2143
Protease Streptomyces griseus, Powder, Type XIV, approx. 4 units/mg solid P5147
Protease from Aspergillus sojae, Type XIX, approx. 0.4 unit/mg solid P7026
Endoproteinase Glu-C Staphylococcus aureus V8, Type XVII-B, Lyophilized powder, 500-1,000 units/mg solid P2922
Protease from Rhizopus sp., Type XVIII, 0.2-0.6 unit/mg solid, pH 3.0, 37°C P5027
Protease Streptomyces griseus, Type XXI, minimum 15 units/mg solid P0652
Proteinase from Aspergillus oryzae, Type XXIII, minimum 3 units/mg solid P4032
Proteinase, bacterial, Type XXIV, Lyophilized powder, 7-14 units/mg solid P8038
Endoproteinase Arg-C murine submaxillary gland, Type XX-S, Lyophilized powder, 250-800 units/mg protein (Lowry) P5171
Calpain II from rabbit skeletal muscle, Lyophilized powder, 15-40 units/mg protein P4533
Protease Bacillus globigii (Bacillus licheniformis), BioChemika, powder, white, 7-15 U/mg 82459
Proteinase Bacillus subtilis var. biotecus A, BioChemika, lyophilized, powder, ~20 U/mg 82490
Proteinase A from baker's yeast (S. cerevisiae), Lyophilized powder, 15-50 units/mg protein P8892
Proteinase K Tritirachium album, Lyophilized powder, minimum 30 units/mg protein (Biuret) P5056
Proteinase K Tritirachium album, Lyophilized powder, 1-3 unit/mg solid P8044
Proteinase K Tritirachium album, Lyophilized powder, minimum 30 units/mg protein P6556
Proteinase K Tritirachium album, Buffered aqueous glycerol solution P4850
Proteinase K Tritirachium album, minimum 500 units/mL, Buffered aqueous glycerol solution P5568
Proteinase K Tritirachium album, minimum 500 units/mL, Buffered aqueous glycerol solution P5568
Proteinase K-Acrylic Beads from Tritirachium album, 2-8 units/g solid P0803
Proteinase K Tritirachium album, BioChemika, for molecular biology, suspension in 10 mM TRIS HCl, pH 7.5, 40% (v/v) glycerol, 1 mM Ca-acetate, ~20000 U/ml 82456
Proteinase K Tritirachium album, BioChemika, powder, white, ³500 U/mg 92905
Proteinase K Tritirachium album, BioChemika, powder, beige, ~290 U/mg 82495
Renin porcine kidney, Lyophilized powder, 0.5-5.0 units/mg protein R2761
Rennet from Mucor miehei, Type II R5876
Rennin calf stomach, minimum 20 units/mg protein R4877
Rennin calf stomach, Lyophilized powder, 50-100 units/mg protein R4879
Rennin Mucor miehei, BioChemika, lyophilized, powder, slightly brown, ~0.1 U/mg 83553
Streptokinase beta;-hemolytic Streptococcus (Lancefield Group C), Buffered aqueous solution, approx. 100,000 units/mg protein S8026
Streptokinase beta;-hemolytic Streptococcus (Lancefield Group C), BioChemika, powder (fine), white, ~2 U/mg 85877
Protease Bacillus globigii (Bacillus licheniformis), BioChemika, ~12 U/mg 85968
Protease Bacillus globigii (Bacillus licheniformis), BioChemika, 95%, powder, light brown, ~6 U/mg 85967
Thermolysin Bacillus thermoproteolyticus rokko, BioChemika, lyophilized, powder, ~40 U/mg 88303
Thrombin bovine plasma, Lyophilized powder, 175-350 NIH units/mg protein (Biuret) T4265
Thrombin bovine plasma, Lyophilized powder, 50-175 NIH units/mg protein (Biuret) T4648
Thrombin bovine plasma, Lyophilized powder, 600-1,500 NIH units/mg protein (Biuret) T6634
Thrombin bovine plasma, Lyophilized powder T7513
Thrombin from human plasma, Lyophilized powder T1063
Thrombin from human plasma, Lyophilized powder, approx. 2,000 NIH units/mg protein (E1%/280 = 18.3) T6884
Thrombin from human plasma, Lyophilized powder, approx. 1,000 NIH units/mg protein (E1%/280 = 18.3) T7009
Thrombin from human plasma, suitable (For routine use in the thrombin time test), Lyophilized powder T8885
Thrombin from human plasma, suitable (For routine use in the thrombin time test.), Lyophilized powder T9135
Thrombin from human plasma, Lyophilized powder, 50-300 NIH units/mg protein T9677
Thrombin from rat plasma, Lyophilized powder, minimum 1,000 units/mg protein (Biuret) T5772
Thrombin from human plasma, BioChemika, lyophilized, powder, white, ~0.8 U/mg 89223
Thrombin bovine plasma, BioChemika, powder, white, ~1 U/mg 89228
Tissue plasminogen activator from human melanoma cells, Lyophilized powder, Fibrinolysis approx. 400,000 IU/mg T7776
Trypsin from bovine pancreas, Lyophilized powder, minimum 7,500 BAEE units/mg solid T4665
Trypsin Agarose, Buffered aqueous suspension, trypsin from bovine lung T1763
Trypsin-Agarose, Buffered aqueous suspension, Trypsin from bovine pancreas T4019
Trypsin from human pancreas, Salt-free, lyophilized powder T6424
Trypsin porcine pancreas, Tablet T7168
Trypsin from bovine pancreas, Essentially salt-free, lyophilized powder, 10,000-15,000 BAEE units/mg protein T8802
Trypsin from bovine pancreas, Type I, approx. 10,000 BAEE units/mg protein T8003
Trypsin porcine pancreas, Type II-S, Lyophilized powder, 1,000-2,000 BAEE units/mg solid T7409
Trypsin porcine pancreas, Type IX-S, Lyophilized powder, 13,000-20,000 BAEE units/mg protein T0303
Trypsin Acetylated from bovine pancreas, synthetic, Type V-S, approx. 10,000 BAEE units/mg protein (Biuret) T6763
Trypsin from bovine pancreas, Type XI, Lyophilized powder, minimum 6,000 BAEE units/mg protein T1005
Trypsin Gadus morhua (Atlantic cod), Type XX-S, 5,000-10,000 units/mg protein T9906
Trypsin from bovine pancreas, USP, minimum 2,500 USP units/mg solid T7309
Trypsin from bovine pancreas, BioChemika, essentially salt free, lyophilized, powder, slightly beige, ~9000 U/mg 93610
Trypsin from bovine pancreas, BioChemika, lyophilized, powder, slightly beige, ~7500 U/mg 93612
Trypsin, DPCC treated, bovine pancreas, BioChemika, lyophilized, powder, off-white, 6000-12000 U/mg 93611
Trypsin, TPCK treated, bovine pancreas, BioChemika, powder (crystalline), ~7500 U/mg 93630
Trypsin porcine pancreas, BioChemika, ³10000 U/mg 93614
Trypsin porcine pancreas, BioChemika, powder, white, ~1500 U/mg 93615
Trypsin porcine pancreas, BioChemika, powder, ~90 U/mg 93613
Trypsinogen from bovine pancreas, Essentially salt-free, lyophilized powder, 10,000-15,000 BAEE units/mg protein (E1%/280, after activation to trypsin) T1143
Trypsinogen from bovine pancreas T1146
Tryptase from human lung, Buffered aqueous solution, minimum 5 units/mg protein T7063
Urokinase from human urine U0633
Protein Sequencing back to top
Aminopeptidase from Aeromonas proteolytica, Lyophilized powder, 50-150 units/mg protein A8200
Aminopeptidase I from Streptomyces griseus, Lyophilized powder, minimum 200 units/mg protein A9934
Carboxypeptidase A-Agarose, Ammonium sulfate suspension, 200-400 units/g agarose, 30°C (One ml gel will yield 6-12 units), carboxypeptidase A from bovine pancreas C1261
Carboxypeptidase A bovine pancreas, Type I, Aqueous suspension, approx. 50 units/mg protein C0261
Carboxypeptidase A bovine pancreas, bovine pancreas, Aqueous suspension, approx. 50 units/mg protein C0386
Carboxypeptidase A, PMSF treated bovine pancreas, bovine pancreas, approx. 50 units/mg protein