Sigma-Aldrich
Biochemicals
Antibiotic Explorer
BioUltra
Biological Detergents
Detection
Enzyme Explorer
BioUltra Proteins
Product Indexes
Cell Signaling Enzymes
Chaetocin
Chymase
Complement Proteins
Cyanobacterial Toxins
Dihydrofolate reductase
Dipeptidyl Peptidase IV
Elafin
Elastase
Matrix Metalloproteinases
Nitric Oxide Metabolism
Superoxide Dismutase
Xenobiotics
Analytical Enzymes
Enzyme Reagents
Key Resources
Lipids
Microparticles
Peptide Explorer
Pharmacopoeia
PolyAmino Acids
 Dihydrofolate Reductase
Enzyme Explorer
Dihydrofolate Reductase
Human, Recombinant
Expressed in E. coli
Product Code D 6566
Primary Accession Number P00374
EC 1.5.1.3
Synonyms: Tetrahydrofolate NADP+ oxidoreductase; DHFR
Purity:
Specific Activity:
Storage/Stability:
Unit definition:
  minimum 80% (SDS-PAGE)
minimum 1 unit per mg protein
This product ships on dry ice and storage at -20°C is recommended.
One unit will convert 1.0 mmole of dihydrofolic acid to tetrahydrofolic acid per minute at pH 7.5 at 22°C.
Dihydrofolate Reductase (DHFR), a key enzyme in thymidine synthesis, catalyzes the NADPH dependent reduction of dihydrofolate (DHF) to tetrahydrofolate (THF) and, at much lower rate, the conversion of folate to THF. The reaction product, THF, is an essential cofactor in the conversion of deoxyuridylate (dUMP) to deoxythymidylate (dTMP) by thymidylate synthetase.1,2 Therefore, DHFR is a critical enzyme in DNA synthesis and has become a target for drug development and cancer therapy. The variations between DHFR from different sources has enabled the development of species selective DHFR inhibitors, such as trimethoprim (antibacterial and antifungal), pyrimethamine (antiprotozoal), and methotrexate, MTX, (antineoplastic, antipsoriatic, and anti-inflammatory).3
Human DHFR is an 186 amino acid protein with an apparent molecular weight of 25 kDa. It is 30% homologous to the E. coli protein and up to 70% homologous to vertebrate proteins.2 The human DHFR gene, as well as other mammalian DHFR genes, overcomes the inhibitory effects of methotrexate by the mechanism of gene amplification or by amino acid mutagenesis.3
This product is supplied as a solution in 10 mM Tris-HCl, pH 8.0, with 1 mM EDTA, 0.5 mM DTT, 5 µM NADPH, protease inhibitors, and 50% glycerol.
Kinetic Properties:
Km5,6
0.16 mM NADPH
0.03 mM 7,8-dihydrofolate
8-methylpterin 0.13 mM
Ki7
Folate 2.6x10-5 mM
Methotrexate 6.1-9x10-9
Related Products:
Inhibitors  
Product Name Product # 
Aminopterin A1784
Methotrexate hydrate A6770
Amethopterin A7019
4-Chloromercuribenzoic acid C5913
Substrate/Cofactor  
Product Name Product # 
Dihydrofolic acid D7006
NADPH N7505
References:
  1. Gready, J.E., Dihydrofolate reductase: binding of substrates and inhibitors and catalytic mechanism. Adv. Pharmacol. Chem., 17, 37-103 (1980).
  2. Blakley, R.L., Eukaryotic dihydrofolate reductase. Adv. Enzymol. Relat. Areas Mol. Biol., 70, 23-101 (1995).
  3. Schweitzer, B.I., et al., Dihydrofolate reductase as a therapeutic target, FASEB J., 4, 2441-2452 (1990).
  4. Hillcoat, B.L., et al., Effect of substrate decomposition on the spectrophotometric assay of dihydrofolate reductase. Anal. Biochem., 21, 178-189 (1967).
  5. T.J. Susten, S.S. Blankenship, D.T. Freisheim, J.H. Delcamp, Purification and characterization of dihydrofolate reductase from methotrexate-resistant human lymphoblastoid cells, Biochemistry, 22,633-639 (1983).
  6. Thibault, V.; Koen, M.J.; Gready, J.E.Enzymic properties of a new mechanism-based substrate for dihydrofolate reductase, Biochemistry, 28, 6042-6049 (1989).
  7. Jarabak, J. Bachur, N.R., A soluble dihydrofolate reductase from human placenta: purification and properties, Arch. Biochem. Biophys., 142, 417-425 (1971).
back to top
back to Enzyme Explorer