Sigma-Aldrich
Biochemicals
Antibiotic Explorer
Biological Buffers
Biological Detergents
Books
Enzyme Explorer
Application Index
Class Index
Inhibitor Index
Substrate Index
Cofactor Index
Lectin Index
Cell Signaling Enzymes
Apoptosis Products
COX Enzymes
Chaetocin
Chymase
Complement Proteins
Cyanobacterial Toxins
Dihydrofolate reductase
Dipeptidyl Peptidase IV
Elafin
Elastase
HIV Protease
Matrix Metalloproteinases
Nitric Oxide Metabolism
Proteasome
Superoxide Dismutase
Xenobiotics
Analytical Enzymes
Enzyme Reagents
Key Resources
Lipids
Peptide Explorer
Pharmacopoeia
PolyAmino Acids
Radiochemicals
Vitamins and Derivatives
 Elafin
Enzyme Explorer
Elafin
Human, Recombinant
Expressed in Saccharomyces cerevisiae
Product Code E 7280
Synonyms: Elastase-specific inhibitor; Skin-Derived Antileukoprotease
Calculated Molecular Masses
    Average Molecular Mass: 5999.20 Da
    Monoisotopic Molecular Mass: 5994.8158 Da
Isoelectric Point (pI): 9.7
Elafin: Inhibitor of Human Leukocyte Elastase & Proteinase
Elafin Peptide Sequence1
Elafin is a peptide found in the human epithelia. It is a reversible, tightly binding inhibitor of human leukocyte elastase and proteinase found in human epithelia 3.
Ki = 1.7 x 10-10 M for human leukocyte elastase2
Ki = 4.2 x 10-10 M for human proteinase 33
Elafin is produced in human skin, lungs, and mammary glands. It may affect the inflammation process and protect cells against destruction by the immune system. Elafin has shown potential applications for the treatment of cardiac infarction and organ rejection, as well as for treatment of seriously injured tissues and inflammatory lung diseases.4
Preparation Instructions
Stock solutions of Elafin may be prepared in water. Subsequent dilutions should be made into aqueous buffers, pH 4.0-8.0.
Storage/Stability
This product ships at ambient temperature and storage at 0 to -20°C is recommended. Stock solutions should be stored at -20°C. Avoid repeated freezing and thawing.
Related Products
Product Name Product # 
Human Leukocyte Elastase E 8140
N-t-BOC-L-Alanine p-Nitropheny l Ester B 5126
References
  1. Wiedow, O., et. al., J. Biol. Chem., 266, 3356 (1991)
  2. Ying, Q.L., et. al., Biochemistry, 32, 1866 (1991)
  3. Ying, Q.L., et. al., J. Respir. Cell. Mol. Biol., 24, 83 (2001)
  4. Sallenave, J.M., Respir. Res., 1, 87-92 (2000)
back to top
back to Enzyme Explorer