Order Center
Biochemicals
Antibiotic Explorer
BioUltra
Biological Detergents
Detection
Enzyme Explorer
BioUltra Proteins
Product Indexes
Cell Signaling Enzymes
Chaetocin
Chymase
Complement Proteins
Cyanobacterial Toxins
Dihydrofolate reductase
Dipeptidyl Peptidase IV
Elafin
Elastase
Matrix Metalloproteinases
Nitric Oxide Metabolism
Superoxide Dismutase
Xenobiotics
Analytical Enzymes
Enzyme Reagents
Key Resources
Lipids
Microparticles
Peptide Explorer
Pharmacopoeia
PolyAmino Acids
 HIV Protease Reagents & Envelope Proteins
Enzyme Explorer
HIV protease (E. C. # EC 3.4.23.16), a virally-encoded aspartic protease belonging to the Retropepsin A2 proteinase family. It is produced following HIV infection of host cells, and processes the gag and gag-pol proteins required during the early replication phase of the virus. Several inhibitors of HIV protease have been developed as successful therapeutic agents. HIV protease is a dimer with a monomer MW of 10,779. Sigma's HIV protease is supplied in the inactive form as a solution in 20 mM HCl. Reactivation occurs when the enzyme is reconstituted in buffers in the pH range of 4 to 7.
  HIV Protease
HIV Protease Inhibitor
HIV Protease Substrates
HIV Proteins
HIV Protease  
Product Name
Product # 
HIV Protease, suitable for cleaving HIV substrate III
P7338
HIV Protease Inhibitor back to top
Product Name
Product # 
a-MAPI, Phe-urea-Arg-Val-Phe-CHO
M9812
HIV Protease Substrates back to top
Product Name
Product # 
Acetyl-Ser-Gln-Asn-Tyr-Pro-Val-Val amide
A0806
Anthranilyl-His-Lys-Ala-Arg-Val-Leu-p-nitro-Phe-Glu-Ala-Nle-Ser amide
A0811
His-Lys-Ala-Arg-Val-Leu-p-nitro-Phe-Glu-Ala-Nle-Ser amide
H5397
HIV Protease Substrate 1
H6660
HIV Protease Substrate III-B, Native Sequence
H5285
HIV Protease Substrate V
H5410
HIV Protease Substrate VI
H5535
HIV Protease Substrate VII
H5660
HIV Protease Substrate VIII
H5785
Anthranilyl-HIV Protease Substrate
H5910
Anthranilyl-HIV Protease Substrate V
H6035
Anthranilyl-HIV Protease Substrate VI
H6160
Lys-Ala-Arg-Val-Nle-p-nitro-Phe-Glu-Ala-Nle amide
L6525
HIV Proteins back to top
Product Name
Product # 
HIV-1 env antigen human, recombinant
H9909
HIV Envelope protein (gp41) Fragment 579-601
H4521
HIV Envelope protein (gp120) Fragment 254-274
H7769
HIV-1 gag
H0160
HIV Envelope protein (gp120) Fragment 308-331
H7894
HIV Envelope protein (gp120) Fragment 421-438
H6153
HIV-1 nef human
H0285
back to top
back to Enzyme Explorer