Sigma-Aldrich
Biochemicals
Antibiotic Explorer
BioUltra
Biological Detergents
Detection
Enzyme Explorer
BioUltra Proteins
Product Indexes
Cell Signaling Enzymes
Chaetocin
Chymase
Complement Proteins
Cyanobacterial Toxins
Dihydrofolate reductase
Dipeptidyl Peptidase IV
Elafin
Elastase
Matrix Metalloproteinases
Nitric Oxide Metabolism
Superoxide Dismutase
Xenobiotics
Analytical Enzymes
Enzyme Reagents
Key Resources
Lipids
Microparticles
Peptide Explorer
Pharmacopoeia
PolyAmino Acids
 Matrix Metalloproteinases
Enzyme Explorer
Access the matrix metalloproteinase you are interested by the links below:
Complete list of Matrix Metalloproteinase products
Feature Article: "Matrix Metalloproteinases and their Inhibitors - Current Status and Future Challenges"
MMP 1, 3, 8, 10, 12, 13, 18, 19, 20, 22, 27 & 28
MMP Alternative Name EC Number Substrates
MMP-1 Collagenase (Type I, interstitial)
3.4.24.7
Collagens (I, II, III, VII, VIII, and X); gelatin; aggrecan; L-selectin; IL-1beta; proteoglycans; entactin; ovostatin; MMP-2; MMP-9
MMP-3 Stromelysin-1, Proteoglycanase
3.4.24.17
Collagens (III, IV, V, and IX); gelatin; aggrecan; perlecan; decorin; laminin; elastin; caesin; osteonectin; ovostatin; entactin; plasminogen; MBP; IL-1beta; MMP-2/TIMP-2; MMP-7; MMP-8; MMP-9; MMP-13
MMP-8 Neutrophil collagenase
3.4.24.34
Collagens (I, II, III, V, VII, VIII, and X); gelatin; aggrecan; fibronectin
MMP-10 Stromelysin-2
3.4.24.22
Collagens (III-V); gelatin; casein; aggrecan; elastin; MMP-1; MMP-8
MMP-12 Macrophage metalloelastase
3.4.24.65
Collagen IV; gelatin; elastin; casein; fibronectin; vitronectin; laminin; entactin; MBP; fibrinogen; fibrin; plasminogen
MMP-13 Collagenase-3
*
Collagens (I, II, III, IV, IX, X, and XIV); gelatin; plasminogen;aggrecan; perlecan; fibronectin; osteonectin; MMP-9
MMP-18 Xenopus Collagenase-4
*
Type I collagen
MMP-19 RASI
*
Type I collagen
MMP-20 Enamelysin
3.4.24
Amelogenin; aggrecan, and cartilage oligomeric matrix protein (COMP)
MMP-22 Chicken MMP (C-MMP)
*
Unknown
MMP-27  
*
Unknown
MMP-28 Epilysin
*
Unknown
back to top
MMP 2
MMP Alternative Name EC Number Substrates
MMP-2 Gelatinase A 72 kDa Gelatinase Type IV Collagenase
3.4.24.24
back to top
MMP 7 & 26
MMP Alternative Name EC Number Substrates
MMP-7 Matrilysin; PUMP
3.4.24.23
Collagens (IV and X); gelatin; aggrecan; decorin; fibronectin; laminin; entactin; elastin; casein; transferrin; plasminogen; MBP; Beta4-intergrin; MMP-1; MMP-2; MMP-9; MMP-9/TIMP-1
MMP-26 Matrilysin-2; Endometase
*
Collagen IV, fibronectin, fibrinogen, gelatin, alpha (1)-proteinase inhibitor
back to top
MMP 9
MMP Alternative Name EC Number Substrates
MMP-9 Gelatinase B
3.4.24.35
Collagens (IV, V, VII, X, and XIV); gelatin; entactin; aggrecan; elastin; fibronectin; osteonectin; plasminogen; MBP; IL-1beta
back to top
MMP 11
MMP Alternative Name EC Number Substrates
MMP-11 Stromelysin-3
3.4.24
Unknown (casein)
back to top
MMP 14, 15, 16, 24
MMP Alternative Name EC Number Substrates
MMP-14 MT1-MMP
3.4.24
Collagens (I-III); gelatin; casein; fibronectin; laminin; vitronectin; entactin; proteoglycans; MMP-2; MMP-13
MMP-15 MT2-MMP
3.4.24
Fibronectin; entactin; laminin; aggrecan; perlecan; MMP-2
MMP-16 MT3-MMP
3.4.24
Collagen III; gelatin; casein; fibronectin; MMP-2
MMP-24 MT5-MMP
*
Fibronectin, but not collagen type I nor laminin
back to top
MMP 17 & 25
MMP Alternative Name EC Number Substrates
MMP-17 MT4-MMP
3.4.24
Unknown
MMP-25 MT6-MMP; leukolysin
*
Pro-gelatinase A
back to top
MMP 21 & 23
MMP Alternative Name EC Number Substrates
MMP-21 Xenopus MMP (X-MMP)
*
Unknown
MMP-23 Cysteine Array Matrix Metalloproteinase (CA-MMP)
*
Unknown
back to top
* EC Number not assigned.