Enzyme Class Index |
 |
Enzyme Explorer |
 |
 |
| |
|
| |
Oxidoreductases |
|
| E.C. # |
Product Name |
Product # |
|
Acting on hydroxyl groups |
|
| 1.1.1.1 |
Alcohol Dehydrogenase from Saccharomyces cerevisiae, suitable for conventional determination of b-NAD, b-NADH, ethanol and acetaldehyde, Lyophilized powder, Solid with buffer salts, 300-500 units/mg protein |
A7011 |
| 1.1.1.1 |
Alcohol Dehydrogenase from Saccharomyces cerevisiae, suitable for recycling micro-assay of b-NAD and b-NADH, Lyophilized powder, Solid with buffer salts, minimum 300 units/mg protein |
A3263 |
| 1.1.1.1 |
Alcohol dehydrogenaseAgarose from Baker's yeast (S. cerevisiae), Lyophilized powder, 900-1,200 units/g agarose, pH 8.8, 30°C |
A2529 |
| 1.1.1.1 |
Alcohol Dehydrogenase equine liver, Lyophilized powder, ³0.5 units/mg protein (TCA/Biuret) |
A9589 |
| 1.1.1.1 |
Alcohol Dehydrogenase from Saccharomyces cerevisiae, BioChemika, powder, off-white, ³250 U/mg |
05635 |
| 1.1.1.1 |
Alcohol Dehydrogenase from Saccharomyces cerevisiae, BioChemika, lyophilized, powder, ~300 U/mg |
05640 |
| 1.1.1.1 |
Alcohol Dehydrogenase equine liver, BioChemika, suspension in 20 mM potassium phosphate buffer, pH 7; ethanol 10%, ampoule, 10% as ethanol, ~30 U/ml |
05645 |
| 1.1.1.1 |
Alcohol Dehydrogenase equine liver, BioChemika, crystalline suspension in 20 mM potassium phosphate buffer, ~3 U/mg protein (~10 mg/ml) |
05646 |
| 1.1.1.1 |
Alcohol Dehydrogenase equine liver, BioChemika, lyophilisate, slightly beige, ³0.25 U/mg |
17233 |
| 1.1.1.1 |
Alcohol Dehydrogenase from Candida boidinii, BioChemika, powder, light brown, ³0.4 U/mg |
91031 |
| 1.1.1.2 |
Alcohol dehydrogenase, NADP+ dependent Thermoanaerobium brockii, Lyophilized powder, 30-90 units/mg protein |
A9287 |
| 1.1.1.2 |
Alcohol Dehydrogenase Thermoanaerobium brockii, Lyophilized powder, 5-15 units/mg protein |
A8435 |
| 1.1.1.2 |
(S)-Aromatic Alcohol Dehydrogenase, NADP+ dependent from Thermoanaerobium sp., Lyophilized powder, 1-5 units/mg solid |
A9685 |
| 1.1.1.2 |
Alcohol dehydrogenase, NADP+ dependent Thermoanaerobium brockii, Lyophilized powder, 30-90 units/mg protein |
A9287 |
| 1.1.1.2 |
Alcohol Dehydrogenase Thermoanaerobium brockii, Lyophilized powder, 5-15 units/mg protein |
A8435 |
| 1.1.1.2 |
(S)-Aromatic Alcohol Dehydrogenase, NADP+ dependent from Thermoanaerobium sp., Lyophilized powder, 1-5 units/mg solid |
A9685 |
| 1.1.1.2 |
Alcohol Dehydrogenase from Lactobacillus kefir, BioChemika, powder, beige, ~0.4 U/mg |
05643 |
| 1.1.1.2 |
Alcohol Dehydrogenase Thermoanaerobium brockii, BioChemika, powder, white, ~35 U/mg |
05655 |
| 1.1.1.6 |
Glycerol Dehydrogenase from Bacillus megaterium, Lyophilized powder, ³15 units/mg solid, pH 8.8 |
G6267 |
| 1.1.1.6 |
Glycerol Dehydrogenase from Cellulomonas sp., Lyophilized powder, 50-125 units/mg protein, pH 10.0 |
G3512 |
| 1.1.1.6 |
Glycerol Dehydrogenase Enterobacter aerogenes, Lyophilized powder, 20-80 units/mg protein, pH 10.0 |
G4783 |
| 1.1.1.6 |
Glycerol Dehydrogenase Geotrichum candidum, BioChemika, ~30 U/g |
49860 |
| 1.1.1.8 |
a-Glycerophosphate Dehydrogenase from rabbit muscle, Type I, Ammonium sulfate suspension, 100-300 units/mg protein |
G6751 |
| 1.1.1.8 |
a-Glycerophosphate Dehydrogenase from rabbit muscle, Type X, Lyophilized powder, 100-300 units/mg protein |
G6880 |
| 1.1.1.8 |
sn-Glycero-3-phosphate Dehydrogenase rabbit muscle, BioChemika, suspension, white, ~270 U/mg protein (~10 mg/ml) |
50013 |
| 1.1.1.8 |
sn-Glycero-3-phosphate Dehydrogenase rabbit muscle, BioChemika, lyophilized, powder, white, 100-250 U/mg |
50015 |
| 1.1.1.10 |
L-Xylulose Reductase from pigeon liver, Ammonium sulfate suspension, 5-15 units/mg protein (BCA) |
X5250 |
| 1.1.1.14 |
Sorbitol Dehydrogenase from sheep liver, Lyophilized powder, minimum 20 units/mg protein |
S3764 |
| 1.1.1.14 |
Sorbitol Dehydrogenase from sheep liver, 20-40 units/mg protein |
S1128 |
| 1.1.1.14 |
SDH Control (Sorbitol Dehydrogenase) |
S8135 |
| 1.1.1.14 |
Sorbitol Dehydrogenase from sheep liver, BioChemika, lyophilized, powder, white, ~6 U/mg |
85535 |
| 1.1.1.18 |
myo-Inositol Dehydrogenase from Enterobacter aerogenes, Lyophilized powder, 25-50 units/mg protein |
I5010 |
| 1.1.1.18 |
myo-Inositol Dehydrogenase from Enterobacter aerogenes, Lyophilized powder, 10-20 units/mg protein |
I0255 |
| 1.1.1.18 |
myo-Inositol Dehydrogenase from Enterobacter aerogenes, 10-20 units/mL, Ammonium sulfate suspension |
I3517 |
| 1.1.1.22 |
Uridine-5¢-diphosphoglucose Dehydrogenase from bovine liver, Type III, Lyophilized powder, 0.02-0.08 units/mg protein |
U5500 |
| 1.1.1.22 |
Uridine-5¢-diphosphoglucose Dehydrogenase from bovine liver, Type VI, Lyophilized powder, 0.3-0.6 units/mg protein |
U7251 |
| 1.1.1.26 |
Glyoxylate reductase from Spinacia oleracea (spinach), Ammonium sulfate suspension, approx. 50 units/mg protein (Biuret) |
G5259 |
| 1.1.1.27 |
L-Lactic Dehydrogenase LDH-5 (M4) Isoenzyme solution from bovine muscle, Type XXVI, Ammonium sulfate suspension, minimum 600 units/mg protein |
L0508 |
| 1.1.1.27 |
L-Lactic Dehydrogenase LDH-5 (M4) Isoenzyme solution from porcine muscle, Type XXXII-S, Ammonium sulfate suspension, approx. 1,000 units/mg protein |
L5762 |
| 1.1.1.27 |
L-Lactic Dehydrogenase LDH-2 (H3M) Isoenzyme from human erythrocytes, Type XX, 98% (homogeneous), Ammonium sulfate suspension, approx. 150 units/mg protein |
L3757 |
| 1.1.1.27 |
L-Lactic Dehydrogenase solution porcine heart, Ammonium sulfate suspension, minimum 400 units/mg protein (Biuret) |
L7525 |
| 1.1.1.27 |
L-Lactic Dehydrogenase Bacillus stearothermophilus, recombinant, expressed in Escherichia coli, Lyophilized powder, minimum 200 units/mg protein, pH 6.0, 30°C (Lowry) |
L3151 |
| 1.1.1.27 |
L-Lactic Dehydrogenase from bovine muscle, Type X, Ammonium sulfate suspension, minimum 600 units/mg protein |
L1378 |
| 1.1.1.27 |
L-Lactic Dehydrogenase isoenzymes LDH-1 (H4) and LDH-2 (H3M) from human erythrocytes, Type XII, Ammonium sulfate suspension, 300-500 units/mg protein |
L3379 |
| 1.1.1.27 |
L-Lactic Dehydrogenase from rabbit muscle, Type XI, Salt-free, lyophilized powder, 700-1,200 units/mg protein |
L1254 |
| 1.1.1.27 |
L-Lactic Dehydrogenase solution from rabbit muscle, Type I, Ammonium sulfate suspension, 40-100 units/mg protein |
L2375 |
| 1.1.1.27 |
L-Lactic Dehydrogenase solution from rabbit muscle, Type II, Ammonium sulfate suspension, 800-1,200 units/mg protein |
L2500 |
| 1.1.1.27 |
L-Lactic Dehydrogenase solution from rabbit muscle, Type V-S, >95%, Ammonium sulfate suspension, minimum 700 units/mg protein |
L5132 |
| 1.1.1.27 |
L-Lactic dehydrogenase Insoluble enzyme attached to beaded agarose from rabbit muscle, Ammonium sulfate suspension, 300-500 units/g agarose |
L8011 |
| 1.1.1.27 |
L-Lactic dehydrogenase Insoluble enzyme attached to polyacrylamide from rabbit muscle, 25-40 units/g solid |
L8253 |
| 1.1.1.27 |
L-Lactic Dehydrogenase LDH-1 (H4) Isoenzyme solution from rabbit heart, Type XXXVI, Ammonium sulfate suspension, 400-600 units/mg protein |
L2889 |
| 1.1.1.27 |
L-Lactic Dehydrogenase from chicken heart, Type VIII, Ammonium sulfate suspension, 500-800 units/mg protein |
L9126 |
| 1.1.1.27 |
L-Lactic Dehydrogenase solution bovine heart, Type III, Ammonium sulfate suspension, 500-700 units/mg protein |
L2625 |
| 1.1.1.27 |
L-Lactic Dehydrogenase solution bovine heart, Type IX, Ammonium sulfate suspension, minimum 400 units/mg protein |
L0377 |
| 1.1.1.27 |
L-Lactic Dehydrogenase solution bovine heart, Type XVII, Buffered aqueous glycerol solution, 400-600 units/mg protein |
L1006 |
| 1.1.1.27 |
L-Lactic Dehydrogenase solution from rabbit muscle, Type XXXIX, Buffered aqueous glycerol solution, 700-1,000 units/mg protein |
L2518 |
| 1.1.1.27 |
L-Lactate Dehydrogenase rabbit muscle, BioChemika, in 2.1 M ammonium sulfate solution, pH ~6.0, suspension, white, ~500 U/mg protein (~10 mg/ml) |
61311 |
| 1.1.1.27 |
L-Lactate Dehydrogenase rabbit muscle, BioChemika, lyophilized, powder, off-white, ~140 U/mg |
61309 |
| 1.1.1.27 |
Lactic Dehydrogenase preparation from bovine heart, BioChemika, off-white, ~250 U/mg protein (~10 mg/ml) |
61310 |
| 1.1.1.28 |
D-Lactic Dehydrogenase Lactobacillus leichmanii, Lyophilized powder, 150-300 units/mg protein |
L3888 |
| 1.1.1.28 |
D-Lactic Dehydrogenase solution from Lactobacillus leichmanii, Ammonium sulfate suspension, 250-500 units/mg protein (Biuret) |
L2011 |
| 1.1.1.28 |
D-Lactic Dehydrogenase Leuconostoc mesenteroides, Ammonium sulfate suspension, 1,000-3,000 units/mg protein (Biuret) |
L2395 |
| 1.1.1.28 |
D-Lactic Dehydrogenase from Staphylococcus epidermidis, Lyophilized powder, minimum 10 units/mg solid |
L9636 |
| 1.1.1.28 |
D-Lactate Dehydrogenase Staphylococci, BioChemika, lyophilisate, white, ~120 U/mg |
17847 |
| 1.1.1.28 |
D-Lactate Dehydrogenase Lactobacillus sp., BioChemika, powder, white, ³400 U/mg |
59023 |
| 1.1.1.28 |
D-Lactic Dehydrogenase Lactobacillus leichmanii, BioChemika, in 3.2 M ammonium sulfate solution, pH ~6.0, suspension, white, ~1000 U/ml |
61306 |
| 1.1.1.30 |
b-Hydroxybutyrate Dehydrogenase Rhodopseudomonas sphaeroides, Type V, Lyophilized powder, 250-750 units/mg protein |
H8509 |
| 1.1.1.30 |
b-Hydroxybutyrate Dehydrogenase from Rhodopseudomonas sphaeroides, Type II, Ammonium sulfate suspension, 5-10 units/mg protein (Biuret) |
H6126 |
| 1.1.1.30 |
b-Hydroxybutyrate Dehydrogenase from Pseudomonas lemoignei, Lyophilized powder, 200-750 units/mg protein |
H9408 |
| 1.1.1.30 |
b-Hydroxybutyrate Dehydrogenase from Rhodopseudomonas sphaeroides, Type III, Ammonium sulfate suspension, minimum 20 units/mg protein (Biuret) |
H4005 |
| 1.1.1.30 |
b-Hydroxybutyrate Dehydrogenase from Pseudomonas lemoignei, 50-100 units/mL, Ammonium sulfate suspension |
H6523 |
| 1.1.1.30 |
b-Hydroxybutyrate Dehydrogenase Rhodopseudomonas sphaeroides, BioChemika, in 3.2 M ammonium sulfate solution, pH ~6.0, suspension, white, 4-10 U/mg protein (~5 mg/ml) |
54975 |
| 1.1.1.37 |
Malic Dehydrogenase from Thermus flavus, Lyophilized powder, minimum 50 units/mg solid |
M7032 |
| 1.1.1.37 |
Malic Dehydrogenase porcine heart, Buffered aqueous glycerol solution, approx. 1,000 units/mg protein (Biuret) |
M2634 |
| 1.1.1.37 |
Malic Dehydrogenase porcine heart, Ammonium sulfate suspension, approx. 400 units/mg protein (Biuret) |
M7383 |
| 1.1.1.37 |
Malic Dehydrogenase from bovine heart, Ammonium sulfate suspension, 2000-4000 units/mg protein (modified Warburg-Christian) |
M9004 |
| 1.1.1.37 |
Malic Dehydrogenase Thermus sp., recombinant, expressed in Escherichia coli, Ammonium sulfate suspension, 100-200 units/mg protein (Lowry) |
M1035 |
| 1.1.1.37 |
Malic Dehydrogenase porcine heart, BioChemika, suspension, slightly beige, ~1200 U/mg protein (~5 mg/ml) |
63170 |
| 1.1.1.42 |
Isocitrate Dehydrogenase preparation porcine heart, Type I |
I1877 |
| 1.1.1.42 |
Isocitric Dehydrogenase (NADP) solution porcine heart, Type IV, Buffered aqueous glycerol solution, 3-20 units/mg protein (Biuretapprox. 0.1% of that activity is obtained with NAD as cofactor.) |
I2002 |
| 1.1.1.42 |
1,3-Indandione, 97% |
I-200-2 |
| 1.1.1.42 |
Isocitrate Dehydrogenase preparation porcine heart, Lyophilized powder, 20-60 units/mg protein (Biuret) |
I2516 |
| 1.1.1.42 |
Isocitrate Dehydrogenase preparation porcine heart, BioChemika, shipped in dry ice, powder, yellow-brown, 4-12 U/mg |
58774 |
| 1.1.1.42 |
Isocitrate Dehydrogenase preparation porcine heart, BioChemika, solution (clear), amber, 3-8 U/mg protein (~12 mg/ml) |
58775 |
| 1.1.1.44 |
6-Phosphogluconic Dehydrogenase from Lactobacillus sp., Lyophilized powder |
P0235 |
| 1.1.1.44 |
6-Phosphogluconic Dehydrogenase from Saccharomyces cerevisiae, Lyophilized powder, 3-6 units/mg solid |
P4553 |
| 1.1.1.44 |
6-Phosphogluconic Dehydrogenase from torula yeast, Type IV, Ammonium sulfate suspension, minimum 20 units/mg protein |
P0507 |
| 1.1.1.44 |
6-Phosphogluconic Dehydrogenase from sheep liver, Lyophilized powder, 2-10 units/mg protein |
P8406 |
| 1.1.1.44 |
6-Phosphogluconic Dehydrogenase from torula yeast, BioChemika, ³15 U/mg protein |
79461 |
| 1.1.1.47 |
Glucose Dehydrogenase Bacillus megaterium, Lyophilized powder, 50-150 units/mg solid |
G7653 |
| 1.1.1.47 |
Glucose Dehydrogenase from Thermoplasma acidophilum, recombinant, expressed in Escherichia coli, Ammonium sulfate suspension, minimum 100 units/mg protein (glucose substrate), minimum 50 units/mg protein (galactose substrate), Solution |
G5909 |
| 1.1.1.47 |
Glucose Dehydrogenase Bacillus megaterium, BioChemika, ~33 U/mg |
49165 |
| 1.1.1.48 |
b-Galactose Dehydrogenase solution from Pseudomonas fluorescens, recombinant, expressed in Escherichia coli, Ammonium sulfate suspension, minimum 50 units/mg protein (Biuret) |
G6637 |
| 1.1.1.49 |
Glucose-6-phosphate DehydrogenaseAgarose from Baker's yeast (S. cerevisiae), Ammonium sulfate suspension, 1,000-1,750 units/g agarose, 30°C |
G5506 |
| 1.1.1.49 |
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, recombinant, expressed in Escherichia coli, Lyophilized powder, 550-1,100 units/mg protein (Biuret) |
G8529 |
| 1.1.1.49 |
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, recombinant, expressed in Escherichia coli, Ammonium sulfate suspension, 550-1,100 units/mg protein (Biuret) |
G8404 |
| 1.1.1.49 |
Glucose-6-phosphate Dehydrogenase from torula yeast, Ammonium sulfate suspension, 300-600 units/mg protein (Biuret) |
G8289 |
| 1.1.1.49 |
Glucose-6-phosphate Dehydrogenase Biotin Labeled from Baker's yeast, Type XV-B, Lyophilized powder, 80-160 units/mg protein |
G3386 |
| 1.1.1.49 |
Glucose-6-phosphate Dehydrogenase from baker's yeast (S. cerevisiae), Type XV, Lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian) |
G6378 |
| 1.1.1.49 |
Glucose-6-phosphate Dehydrogenase from baker's yeast (S. cerevisiae), Type IX, Lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian) |
G4134 |
| 1.1.1.49 |
Glucose-6-phosphate Dehydrogenase from baker's yeast (S. cerevisiae), Type VII, Ammonium sulfate suspension, 300-400 units/mg protein (modified Warburg-Christian) |
G7877 |
| 1.1.1.49 |
Glucose-6-phosphate Dehydrogenase from baker's yeast (S. cerevisiae), Type V, Dry ammonium sulfate cake, ³150 units/mg protein (modified Warburg-Christian) |
G7750 |
| 1.1.1.49 |
Glucose-6-phosphate Dehydrogenase from torula yeast, Lyophilized powder, 200-600 units/mg protein (BiuretContains approx. 20% sodium citrate based on dry weight.) |
G8164 |
| 1.1.1.49 |
Glucose-6-phosphate Dehydrogenase from yeast, BioChemika, lyophilized, powder, off-white, ~160 U/mg |
49273 |
| 1.1.1.49 |
Glucose-6-phosphate Dehydrogenase from yeast, BioChemika, standardized with bovine serum albumin, crystalline suspension in 3.2 M ammonium sulfate solution, pH ~6, ~120 U/mg protein (~5 mg/ml) |
49270 |
| 1.1.1.49 |
Glucose-6-phosphate Dehydrogenase from baker's yeast (S. cerevisiae), BioChemika, crystalline suspension in 3.2 M ammonium sulfate solution, pH ~6, ~240 U/mg protein (~1 mg/ml) |
49271 |
| 1.1.1.49 |
Glucose-6-phosphate Dehydrogenase from baker's yeast (S. cerevisiae), BioChemika, lyophilized, powder, white, ~210 U/mg |
49272 |
| 1.1.1.49 |
Glucose-6-phosphate Dehydrogenase from torula yeast, BioChemika, suspension, white, ~180 U/mg protein (~1 mg/ml) |
49279 |
| 1.1.1.49 |
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, BioChemika, lyophilized, powder, slightly beige, ~170 U/mg |
49275 |
| 1.1.1.50 |
3a-Hydroxysteroid Dehydrogenase preparation from Pseudomonas testosteroni, Lyophilized powder, 20-50 units/mg protein |
H1506 |
| 1.1.1.50 |
3a-Hydroxysteroid Dehydrogenase preparation from Pseudomonas testosteroni, 50-100 units/mL, Ammonium sulfate suspension |
H6398 |
| 1.1.1.50 |
3a-Hydroxysteroid Dehydrogenase from Pseudomonas testosteroni, BioChemika, lyophilized, powder, white, ~9 U/mg |
56460 |
| 1.1.1.67 |
Mannitol Dehydrogenase from Leuconostoc mesenteroides |
M9532 |
| 1.1.1.95 |
3-Phosphoglycerate Dehydrogenase from chicken liver, Lyophilized powder, 1.5-7.0 units/mg protein (modified Warburg-Christian) |
P4579 |
| 1.1.1.119 |
Glucose Dehydrogenase, NADP dependent from Cryptococcus uniguttulatus, Lyophilized powder, minimum 10 units/mg solid |
G0649 |
| 1.1.1.176 |
12a-Hydroxysteroid Dehydrogenase from Bacillus sphaericus, Lyophilized powder, 150-350 units/mg protein (Lowry) |
H2265 |
| 1.1.1.176 |
12a-Hydroxysteroid Dehydrogenase Clostridium sp., BioChemika, solution, yellow, ³18 U/ml |
37762 |
| 1.1.1.205 |
IMPDH |
I1782 |
| 1.1.1.233 |
N-Acylmannosamine 1-Dehydrogenase from Pseudomonas sp., Lyophilized powder, minimum 45 units/mg protein |
A7561 |
| 1.1.2.3 |
L-Lactic Dehydrogenase (cytochrome) from baker's yeast (S. cerevisiae), Type IV-SS, Ammonium sulfate suspension, 0.1-0.6 units/mg protein (Biuret) |
L4506 |
| 1.1.3.4 |
Glucose Oxidase Aspergillus niger, Type V-S, approx. 1,000 units/mL (without added oxygen), Buffered aqueous solution |
G6891 |
| 1.1.3.4 |
Glucose Oxidase Aspergillus niger, 2,000-10,000 units/g solid (without added oxygen) |
G6766 |
| 1.1.3.4 |
Glucose Oxidase Aspergillus niger, Type VII, Lyophilized powder, 100,000-200,000 units/g solid (without added oxygen) |
G2133 |
| 1.1.3.4 |
Glucose Oxidase Aspergillus niger, Type II, 15,000-25,000 units/g solid (without added oxygen) |
G6125 |
| 1.1.3.4 |
Glucose Oxidase Aspergillus niger, Type X-S, Lyophilized powder, 100,000-250,000 units/g solid (without added oxygen) |
G7141 |
| 1.1.3.4 |
Glucose Oxidase Aspergillus niger, Buffered aqueous solution, minimum 200 units/mg protein (Biuret) |
G9010 |
| 1.1.3.4 |
Glucose Oxidase Aspergillus niger, Type II-S, 25,000-50,000 units/g solid (without added oxygen) |
G6641 |
| 1.1.3.4 |
Glucose Oxidase Aspergillus niger, Type VII-S, Lyophilized powder, 100,000-250,000 units/g solid (without added oxygen) |
G7016 |
| 1.1.3.4 |
Glucose Oxidase Aspergillus niger, BioChemika, solution (clear), yellow, ~1000 U/ml |
49177 |
| 1.1.3.4 |
Glucose Oxidase Aspergillus niger, BioChemika, lyophilized, powder, ~200 U/mg |
49180 |
| 1.1.3.4 |
Glucose Oxidase Aspergillus niger, BioChemika, powder, yellow, ~150 U/mg |
49182 |
| 1.1.3.4 |
Glucose Oxidase Aspergillus niger, BioChemika, powder, yellow, ~130 U/mg |
49181 |
| 1.1.3.4 |
Glucose Oxidase Aspergillus niger, BioChemika, lyophilized, powder, ~25 U/mg |
49178 |
| 1.1.3.6 |
Cholesterol Oxidase from Pseudomonas fluorescens, Lyophilized powder, 10-50 units/mg protein |
C7149 |
| 1.1.3.6 |
Cholesterol Oxidase microbial, recombinant, expressed in Escherichia coli, Lyophilized powder |
C1235 |
| 1.1.3.6 |
Cholesterol Oxidase from Streptomyces sp., Lyophilized powder, minimum 25 units/mg protein |
C8649 |
| 1.1.3.6 |
Cholesterol Oxidase from Cellulomonas sp., Buffered aqueous solution, 20-60 units/mg protein (Biuret) |
C5421 |
| 1.1.3.6 |
Cholesterol Oxidase Nocardia erythropolis, BioChemika, in 1 M ammonium sulfate solution, pH 6, solution (slightly hazy), pale yellow, ³15 U/ml |
26746 |
| 1.1.3.6 |
Cholesterol Oxidase from Pseudomonas sp., BioChemika, powder, light yellow, >1 U/mg |
26747 |
| 1.1.3.9 |
Galactose Oxidase from Dactylium dendroides, Lyophilized powder, 500-1,500 units/mg protein |
G7907 |
| 1.1.3.9 |
Galactose Oxidase from Dactylium dendroides, Lyophilized powder, minimum 3,000 units/g solid |
G7400 |
| 1.1.3.10 |
Pyranose Oxidase from Cariolus sp., recombinant, expressed in Escherichia coli, 2.7 units/mg protein |
P4234 |
| 1.1.3.13 |
Alcohol Oxidase Candida boidinii, Lyophilized powder, 5-15 units/mg protein |
A6941 |
| 1.1.3.13 |
Alcohol Oxidase from Hansenula sp., Vacuum-dried powder, 20-40 units/mg protein (Bradford) |
A0438 |
| 1.1.3.13 |
Alcohol Oxidase solution from Pichia pastoris, BioChemika, solution, red to deep red, ³400 U/ml |
92705 |
| 1.1.3.15 |
Glycolate Oxidase Spinacia oleracea (spinach), Ammonium sulfate suspension, 3-12 units/mg protein, pH 7.8 (Biuret) |
G8260 |
| 1.1.3.17 |
Choline Oxidase from Alcaligenes sp., Lyophilized powder, approx. 10 units/mg solid |
C5896 |
| 1.1.3.17 |
Choline Oxidase from Alcaligenes sp., BioChemika, powder, yellow, 9-18 U/mg |
26986 |
| 1.1.3.17 |
Choline Oxidase from Arthrobacter globiformis, Lyophilized powder, 8-20 units/mg solid |
C4405 |
| 1.1.3.21 |
Glycerol 3-phosphate Oxidase from Pediococcus sp., Lyophilized powder, 40-80 units/mg solid, pH 8.1 |
G9637 |
| 1.1.3.21 |
Glycerol 3-phosphate Oxidase from Streptococcus thermophilus, Lyophilized powder, minimum 10 units/mg solid, pH 7.0 |
G4388 |
| 1.1.3.21 |
Glycerol 3-phosphate Oxidase from Aerococcus viridans, Lyophilized powder, minimum 70 units/mg solid, pH 8.1 |
G9888 |
| 1.1.3.21 |
Glycerol 3-phosphate Oxidase from Aerococcus viridans, BioChemika, powder, yellow, ~40 U/mg |
50030 |
| 1.1.3.22 |
Xanthine Oxidase from buttermilk, Lyophilized powder, 0.4-1.0 units/mg protein |
X4376 |
| 1.1.3.22 |
Xanthine Oxidase from buttermilk, Grade I, Ammonium sulfate suspension, approx. 0.5 units/mg protein |
X1875 |
| 1.1.3.22 |
Xanthine Oxidase from buttermilk, Grade III, Ammonium sulfate suspension, 1-2 units/mg protein |
X4500 |
| 1.1.3.22 |
Xanthine Oxidase microbial, Lyophilized powder, minimum 7 units/mg solid |
X2252 |
| 1.1.3.22 |
Xanthine Oxidase from buttermilk, Grade IV, Ammonium sulfate suspension, 0.1-0.4 units/mg protein |
X4875 |
| 1.1.3.22 |
Xanthine Oxidase from buttermilk, BioChemika, lyophilized, powder, brown, ~0.4 U/mg |
95493 |
| 1.1.3.29 |
N-Acylhexosamine Oxidase from Pseudomonas sp., Solution, minimum 20 units/mg protein |
A0186 |
| 1.1.99.11 |
D-Fructose Dehydrogenase from Gluconobacter industrius, Lyophilized powder, 400-1,200 units/mg protein |
F5152 |
| 1.1.99.11 |
D-Fructose Dehydrogenase from Gluconobacter industrius, Lyophilized powder, minimum 20 units/mg solid |
F4892 |
| 1.1.99.11 |
D-Fructose Dehydrogenase from Gluconobacter industrius, BioChemika, powder, light red, ³20 U/mg |
70052 |
|
Acting on aldehydes or oxo groups |
|
| 1.2.1.1 |
Formaldehyde Dehydrogenase (glutathione-dependent) from Candida boidinii, Lyophilized powder, 7-20 units/mg protein |
F4136 |
| 1.2.1.1 |
Formaldehyde Dehydrogenase (glutathione-dependent) from Candida boidinii, BioChemika, ~2.5 U/mg |
47584 |
| 1.2.1.1 |
Formate Dehydrogenase Pseudomonas spec., recombinant from E. coli, BioChemika, solution, light yellow, ~100 U/ml |
79900 |
| 1.2.1.2 |
Formate Dehydrogenase Candida boidinii, Lyophilized powder, 1.5-4.5 units/mg protein |
F5632 |
| 1.2.1.2 |
Formate Dehydrogenase Candida boidinii, Lyophilized powder, 5-15 units/mg protein |
F8649 |
| 1.2.1.2 |
Formate Dehydrogenase Pseudomonas spec., recombinant mutante to 79900 from E. coli, BioChemika, solution, light yellow, >150 U/ml |
75274 |
| 1.2.1.2 |
Formate Dehydrogenase Pseudomonas spec., recombinant mutante to 79900 NADP-dependant from E. coli from E. coli, BioChemika, solution, light yellow, ~50 U/ml |
79331 |
| 1.2.1.2 |
Formate Dehydrogenase Candida boidinii, BioChemika, powder, 0.3-0.6 U/mg |
47709 |
| 1.2.1.2 |
Formate Dehydrogenase Candida boidinii, BioChemika, liquid (clear), brown, ~50 U/ml |
47753 |
| 1.2.1.5 |
Aldehyde Dehydrogenase, potassium-activated baker's yeast (S. cerevisiae), Lyophilized powder, 2-10 units/mg protein |
A6338 |
| 1.2.1.12 |
Glyceraldehyde-3-phosphate Dehydrogenase human erythrocytes, Lyophilized powder, 50-150 units/mg protein |
G6019 |
| 1.2.1.12 |
Glyceraldehyde-3-phosphate Dehydrogenase from chicken muscle, Lyophilized powder, 40-100 units/mg protein |
G9263 |
| 1.2.1.12 |
Glyceraldehyde-3-phosphate Dehydrogenase from rabbit muscle, Ammonium sulfate suspension, 80-120 units/mg protein (Biuret) |
G0763 |
| 1.2.1.12 |
Glyceraldehyde-3-phosphate Dehydrogenase from rabbit muscle, Lyophilized powder, approx. 80 units/mg protein |
G2267 |
| 1.2.1.12 |
Glyceraldehyde-3-phosphate Dehydrogenase from Baker's yeast (S. cerevisiae), Lyophilized powder, 70-140 units/mg protein |
G5537 |
| 1.2.1.12 |
Glyceraldehyde-3-phosphate Dehydrogenase Agarose from Baker's yeast (S. cerevisiae), Lyophilized powder, 50-150 units/g dry agarose, pH 7.6, 30°C |
G0389 |
| 1.2.1.12 |
Glyceraldehyde-3-phosphate Dehydrogenase from rabbit muscle, BioChemika, suspension, white, ³50 U/mg protein (~10 mg/ml) |
49830 |
| 1.2.1.16 |
Succinic-Semialdehyde Dehydrogenase from Pseudomonas fluorescens, Lyophilized powder, 10-30 units/mg protein |
S3907 |
| 1.2.1.46 |
Formaldehyde Dehydrogenase from Pseudomonas putida, Lyophilized powder, 1-6 units/mg solid |
F1879 |
| 1.2.3.3 |
Pyruvate Oxidase bacterial, Lyophilized powder, minimum 1.5 units/mg |
P4591 |
| 1.2.3.3 |
Pyruvate Oxidase from Aerococcus sp., Lyophilized powder, minimum 35 units/mg protein (Biuret) |
P4105 |
| 1.2.3.4 |
Oxalate Oxidase from barley seedlings, Lyophilized powder, 0.5-1.5 units/mg solid |
O4127 |
| 1.2.4.2 |
a-Ketoglutarate Dehydrogenase from porcine heart, Buffered aqueous glycerol solution, 0.1-1 units/mg protein (Lowry) |
K1502 |
|
Acting on CH-CH |
|
| 1.3.3.5 |
Bilirubin Oxidase from Myrothecium verrucaria, Lyophilized powder, 15-65 units/mg protein |
B0390 |
| 1.3.3.6 |
Acyl coenzyme A oxidase from Arthrobacter sp., Lyophilized powder, 10-40 units/mg protein |
A2167 |
| 1.3.3.6 |
Acyl coenzyme A oxidase from Candida sp., Lyophilized powder, 10-30 units/mg protein |
A2903 |
|
Acting on CH-NH2 |
|
| 1.4.1.1 |
L-Alanine Dehydrogenase Bacillus subtilis, Buffered aqueous glycerol solution, approx. 30 units/mg protein (Lowry) |
A7653 |
| 1.4.1.1 |
L-Alanine Dehydrogenase Bacillus subtilis, Ammonium sulfate suspension, approx. 40 units/mg protein (Lowry) |
A7189 |
| 1.4.1.1 |
L-Alanine Dehydrogenase Bacillus subtilis, BioChemika, in 2.4 M ammonium sulfate solution, pH 7.0, suspension, white, ³30 U/mg protein (~10 mg/ml) |
05192 |
| 1.4.1.3 |
L-Glutamic Dehydrogenase solution from bovine liver, Type I, Ammonium sulfate suspension, approx. 40 units/mg protein |
G2501 |
| 1.4.1.3 |
L-Glutamic Dehydrogenase solution from bovine liver, Type II, Buffered aqueous glycerol solution, approx. 40 units/mg protein |
G2626 |
| 1.4.1.3 |
Glutamate Dehydrogenase bovine liver, Type III, Lyophilized powder, approx. 25 units/mg protein |
G7882 |
| 1.4.1.3 |
Glutamate Dehydrogenase bovine liver, BioChemika, solution (viscous), amber, ~1600 U/ml |
49390 |
| 1.4.1.3 |
Glutamate Dehydrogenase bovine liver, BioChemika, lyophilized, powder, >30 U/mg |
49392 |
| 1.4.1.4 |
L-Glutamic Dehydrogenase (NADP) from Proteus sp., Buffered aqueous solution, minimum 400 units/mg protein (Biuret) |
G4387 |
| 1.4.1.4 |
L-Glutamic Dehydrogenase (NADP) from Candida utilis, Lyophilized powder, 50-200 units/mg protein |
| | |