| |
Lyases |
|
| E.C. # |
Product Name |
Product # |
|
C-C bonds |
|
| 4.1.1.23 |
Orotidine-5¢-monophosphate decarboxylase baker's yeast (S. cerevisiae), Lyophilized powder, 30-65 units/mg protein (Biuret) |
O9251 |
| 4.1.1.3 |
Oxaloacetate decarboxylase Pseudomonas sp., 100-350 units/mg solid |
O4878 |
| 4.1.1.3 |
Oxaloacetate decarboxylase Pseudomonas sp., BioChemika, lyophilized, powder, slightly brown, ~140 U/mg |
75657 |
| 4.1.1.5 |
a-Acetolactate Decarboxylase Bacillus subtilis, Lyophilized powder, approx. 60 units/mg solid |
A7206 |
| 4.1.1.5 |
a-Acetolactate Decarboxylase Bacillus subtilis, BioChemika, Lyophilized powder, brown, ~60 U/mg |
00550 |
| 4.1.1.15 |
L-Glutamic Acid Decarboxylase Escherichia coli, Type II, Partially purified powder, 2.5-5.5 units/mg solid |
G2126 |
| 4.1.1.15 |
L-Glutamic Acid Decarboxylase Clostridium perfringens (C. welchii), Type III, Acetone powder, approx. 0.02 unit/mg solid |
G2251 |
| 4.1.1.15 |
L-Glutamic Acid Decarboxylase Clostridium perfringens (C. welchii), Type IV, Partially purified powder, 0.8-1.5 units/mg solid |
G2376 |
| 4.1.1.18 |
L-Lysine Decarboxylase Bacterium cadaveris, Type V, Partially purified powder, 1-3 units/mg protein (Biuret) |
L0882 |
| 4.1.1.18 |
L-Lysine Decarboxylase Bacterium cadaveris, Type III, approx. 0.01 unit/mg solid |
L6251 |
| 4.1.1.18 |
L-Lysine Decarboxylase Bacterium cadaveris, Type I, approx. 0.08 unit/mg solid |
L6126 |
| 4.1.1.18 |
L-Lysine Decarboxylase Bacterium cadaveris, BioChemika, crude acetone extract, ³0.1 U/mg substance |
62890 |
| 4.1.1.19 |
L-Arginine Decarboxylase from Escherichia coli, ATCC 10787, Type II, Lyophilized powder, 5-15 units/mg protein |
A5381 |
| 4.1.1.22 |
L-Histidine Decarboxylase Clostridium perfringens (C. welchii), Type I, Crude acetone powder, 0.01-0.03 unit/mg solid |
H8250 |
| 4.1.1.22 |
L-Histidine Decarboxylase Lactobacillus 30a, Type III-S, Crude acetone powder, 0.25-0.5 unit/mg solid |
H3266 |
| 4.1.1.25 |
L-Tyrosine Decarboxylase Streptococcus faecalis, minimum 0.01 unit/mg solid |
T7677 |
| 4.1.1.25 |
L-Tyrosine Decarboxylase apoenzyme Streptococcus faecalis, <0.005 unit/mg solid (without pyridoxal 5-phosphate), minimum 0.05 unit/mg solid (with excess pyridoxal 5-phosphate) |
T7927 |
| 4.1.1.25 |
L-Tyrosine Decarboxylase Streptococcus faecalis, BioChemika, powder, beige, ³0.2 U/mg |
93904 |
| 4.1.1.31 |
Phospho(enol)pyruvate Carboxylase corn, Ammonium sulfate suspension, minimum 1 units/mg protein |
P2023 |
| 4.1.1.31 |
Phospho(enol)pyruvate Carboxylase corn, BioChemika, powder, white, ~1 U/mg |
79414 |
| 4.1.1.39 |
D-Ribulose 1,5-diphosphate Carboxylase from spinach, Partially purified powder, 0.01-0.1 unit/mg solid |
R8000 |
| 4.1.1.53 |
L-Phenylalanine Decarboxylase from Streptococcus faecalis, Dried cells, minimum 5 units/g solid |
P9957 |
| 4.1.1.53 |
L-Phenylalanine Decarboxylase from Streptococcus faecalis, BioChemika, powder, yellow-brown, 7 U/mg |
78087 |
| 4.1.2.4 |
2-Deoxyribose-5-phosphate Aldolase from Lactobacillus plantarum, BioChemika, powder, slightly yellow, ~15 U/g |
41228 |
| 4.1.2.5 |
L-Threonine Aldolase Candida humicola, BioChemika, Lyophilisate, slightly beige, ~10 U/g |
53806 |
| 4.1.2.5 |
L-Threonine Aldolase from Pseudomonas putida, BioChemika, Lyophilisate, slightly yellow, ~0.2 U/mg |
96586 |
| 4.1.2.10 |
Mandelonitrile Lyase almonds, Ammonium sulfate suspension, 80-240 units/mg protein (Biuret) |
M6782 |
| 4.1.2.13 |
Aldolase from rabbit muscle, Lyophilized powder, approx. 10 units/mg protein |
A2714 |
| 4.1.2.13 |
Aldolasepolyacrylamide from rabbit muscle, 10-20 units/g solid, 30°C |
A1386 |
| 4.1.2.13 |
Aldolase from spinach, Lyophilized powder, 0.3-1.0 units/mg protein |
A9329 |
| 4.1.2.13 |
Aldolase from rabbit muscle, Ammonium sulfate suspension, 10-20 units/mg protein |
A8811 |
| 4.1.2.13 |
Aldolase from Staphylococcus aureus, Type XII, Lyophilized powder, approx. 20 units/mg protein |
A2548 |
| 4.1.2.13 |
Aldolase from rabbit muscle, BioChemika, powder, white, 15-20 U/mg |
05518 |
| 4.1.2.13 |
Aldolase from rabbit muscle, BioChemika, crystalline suspension in 3.2 M ammonium sulfate solution, pH ~6, ~9 U/mg protein (~10 mg/ml) |
05520 |
| 4.1.2.13 |
Aldolase from Staphylococcus aureus, BioChemika, powder, white, ³10 U/mg |
05522 |
| 4.1.2.13 |
Aldolase from Thermus aquaticus, BioChemika, lyophilisate, yellow, ~5 U/g |
05525 |
| 4.1.2.13 |
Fructose-1,6-bisphosphate Aldolase from Bacillus subtilis, BioChemika, lyophilisate, slightly beige, ~30 U/g |
80753 |
| 4.1.2.13 |
Fructose-1,6-bisphosphate Aldolase Staphylococcus carnosus, BioChemika, lyophilisate, slightly beige, ~0.4 U/mg |
94864 |
| 4.1.2.37 |
(S)-Oxynitrilase from Manihot esculenta (Maniok), BioChemika, solution (turbid), ³3000 U/ml |
71811 |
| 4.1.3.1 |
Isocitrate lyase from Bacillus stearothermophilus, Ammonium sulfate suspension, minimum 0.5 units/mg protein (Biuret) |
I6754 |
| 4.1.3.2 |
Malate Synthase from Bacillus stearothermophilus, Ammonium sulfate suspension, minimum 25 units/mg protein (Biuret) |
M0920 |
| 4.1.3.3 |
N-Acetylneuraminic acid Aldolase Escherichia coli, Lyophilized powder, minimum 20 units/mg protein, pH 7.7 (Biuret) |
A6680 |
| 4.1.3.3 |
N-Acetylneuraminic acid Aldolase Escherichia coli, BioChemika, powder, slightly yellow, ³15 U/mg |
47153 |
| 4.1.3.6 |
Citrate Lyase from Enterobacter aerogenes, Lyophilized powder, minimum 0.2 unit/mg solid |
C0897 |
| 4.1.3.6 |
Citrate Lyase from Enterobacter aerogenes, BioChemika, powder, slightly beige, ~0.3 U/mg |
27457 |
| 4.1.3.7 |
Citrate Synthase from Thermoplasma acidophilum, recombinant, expressed in Escherichia coli, Ammonium sulfate suspension, minimum 10 units/mg protein (Biuret) |
C9454 |
| 4.1.3.7 |
Citrate Synthase from porcine heart, Ammonium sulfate suspension, 100-200 units/mg protein (E1%/280) |
C3260 |
| 4.1.3.7 |
Citrate Synthase from pigeon breast muscle, Ammonium sulfate suspension, 80-150 units/mg protein (modified Warburg-Christian) |
C4140 |
| 4.1.3.16 |
4-Hydroxy-2-oxoglutarate Aldolase from E. coli, BioChemika, lyophilisate, white, ~0.1 U/mg |
59892 |
| 4.1.99.1 |
Tryptophanase from Escherichia coli, 15-40 units/mg solid |
T0754 |
| 4.1.99.1 |
Tryptophanase from microorganisms, BioChemika, solution, yellow, ³2.5 U/mg protein (~100 mg/ml) |
52384 |
| 4.1.99.2 |
b-Tyrosinase, BioChemika, solution, deep yellow, ³25 U/mg protein (~6 mg/ml) |
40522 |
|
C-O bonds |
|
| 4.2.1.1 |
Carbonic Anhydrase Isozyme II from human erythrocytes, minimum 80%, Lyophilized powder, minimum 3,000 W-A units/mg protein |
C6165 |
| 4.2.1.1 |
Carbonic Anhydrase Isozyme II from bovine erythrocytes, Lyophilized powder, minimum 2,000 W-A units/mg protein |
C3640 |
| 4.2.1.1 |
Carbonic Anhydrase I human erythrocytes, Lyophilized powder, 100-500 W-A units/mg protein |
C4396 |
| 4.2.1.1 |
Carbonic Anhydrase from rabbit erythrocytes, Lyophilized powder, 1,000-4,000 W-A units/mg protein |
C1266 |
| 4.2.1.1 |
Carbonic Anhydrase bovine erythrocytes, Lyophilized powder, minimum 2,500 W-A units/mg protein |
C3934 |
| 4.2.1.1 |
Carbonic Anhydrase II bovine erythrocytes, Lyophilized powder, minimum 3,000 W-A units/mg protein |
C2522 |
| 4.2.1.1 |
Carbonic Anhydrase bovine erythrocytes, BioChemika, powder, white, >2000 U/mg |
21803 |
| 4.2.1.1 |
Carbonic Anhydrase Isozyme II from bovine erythrocytes, BioChemika, powder, white, ~1800 U/mg |
21807 |
| 4.2.2.1 |
Hyaluronidase Streptomyces hyalurolyticus, BioChemika, lyophilisate, white to slightly brown, ~0.1 U/vial |
53725 |
| 4.2.1.2 |
Fumarase porcine heart, Ammonium sulfate suspension, 300-500 units/mg protein (Biuret) |
F1757 |
| 4.2.1.2 |
Fumarase porcine heart, Ammonium sulfate suspension, 300-500 units/mg protein (Biuret) |
F1757 |
| 4.2.1.3 |
Aconitase from porcine heart, suitable for assay of citrate when used in conjunction with isocitric dehydrogenase and NADP |
A5384 |
| 4.2.1.11 |
Enolase from baker's yeast (S. cerevisiae), Lyophilized powder, minimum 60 units/mg protein |
E6126 |
| 4.2.1.11 |
Enolase solution from rabbit muscle, Ammonium sulfate suspension, 25-35 units/mg protein (Biuret) |
E0379 |
| 4.2.1.17 |
Crotonase from bovine liver, Lyophilized powder, 500-1000 units/mg protein |
C6073 |
| 4.2.1.24 |
d-Aminolevulinate Dehydratase from bovine liver, Lyophilized powder, 0.5-4.0 units/mg protein |
A0442 |
| 4.2.2.- |
Chondroitinase C from Flavobacterium heparinum, Lyophilized powder, minimum 200 units/mg solid |
C0954 |
| 4.2.2.1 |
Hyaluronidase Streptomyces hyalurolyticus, Lyophilized powder |
H1136 |
| 4.2.2.3 |
Alginate Lyase from Flavobacterium sp., Powder |
A6973 |
| 4.2.2.4 |
Chondroitinase ABC from Proteus vulgaris, Lyophilized powder, 50-250 units/mg protein (using chondroitin sulfate C as substrate) |
C3667 |
| 4.2.2.4 |
Chondroitinase ABC from Proteus vulgaris, Lyophilized powder, 0.3-3 units/mg solid (using chondroitin sulfate C as substrate) |
C2905 |
| 4.2.2.4 |
Chondroitinase ABC from Proteus vulgaris, BioChemika, lyophilized, powder, slightly beige, 0.5-2 U/mg |
27038 |
| 4.2.2.5 |
Chondroitinase AC from Arthrobacter aurescens |
C2262 |
| 4.2.2.5 |
Chondroitinase AC from Flavobacterium heparinum, Lyophilized powder, 0.5-1.5 units/mg solid (using chondroitin sulfate A as substrate, Also cleaves chondroitin sulfate C) |
C2780 |
| 4.2.2.7 |
Heparinase I from Flavobacterium heparinum, Lyophilized powder stabilized with approx. 25% bovine serum albumin., 200-600 unit/mg solid |
H2519 |
| 4.2.2.7 |
Heparinase I from Flavobacterium heparinum, BioChemika, crystals (fine), white, ~1 U/mg |
51534 |
| 4.2.2.7 |
Heparinase I from Flavobacterium heparinum, BioChemika, lyophilized, powder, white, ~0.4 U/mg |
51539 |
| 4.2.2.8 |
Heparinase III from Flavobacterium heparinum, 200-600 unit/mg solid, Lyophilized powder stabilized with approx. 25% bovine serum albumin |
H8891 |
| 4.2.2.10 |
Pectin Lyase acidic enzyme from Aspergillus niger, Buffered aqueous glycerol solution, 50-150 units/mg protein, pH 5.0 (Lowry, assayed in 100 mM citrate-phosphate buffer) |
P7052 |
|
C-N bonds |
|
| 4.3.1.3 |
Histidase from Pseudomonas fluorescens, Dried cells, 10-20 units/mg |
H7500 |
| 4.3.1.5 |
Phenylalanine Ammonia-Lyase from potato, Lyophilized powder, 0.001-0.01 units/mg protein |
P7774 |
| 4.3.1.5 |
Phenylalanine Ammonia-Lyase Rhodotorula glutinis, Grade I, Buffered aqueous glycerol solution, 0.8-2 units/mg protein |
P1016 |
| 4.3.1.5 |
Phenylalanine Ammonia-Lyase Rhodotorula glutinis, Grade II, Buffered aqueous glycerol solution, 0.2-0.8 units/mg protein |
P9519 |
|
C-S bonds |
|
| 4.4.1.5 |
Glyoxalase I from Saccharomyces cerevisiae, Grade IV, Buffered aqueous glycerol solution, 400-800 units/mg protein |
G4252 |
| 4.3.1.5 |
Phenylalanine Ammonia-Lyase Rhodotorula glutinis, BioChemika, in 60% glycerol, 3 mM TRIS HCl, pH 7.5 and 0.5 M ammonium sulfate, clear, colorless, >0.4 U/mg protein (~8 mg/ml) |
78085 |
| 4.3.1.5 |
Phenylalanine Ammonia-Lyase Rhodotorula glutinis, BioChemika, lyophilized, powder, ~0.2 U/mg |
78084 |
| 4.4.1.11 |
L-Methionine g-lyase from Pseudomonas putida, Lyophilized powder |
M4545 |
| 4.6.1.1 |
Adenylate Cyclase Escherichia coli, Buffered aqueous glycerol solution, 50-100 units/mg protein (Biuret) |
A0951 |
| | |
back to top |
Oxidoreductases | Transferases | Hydrolases (Acting on esters) | Hydrolases (Glycosidases) | Hydrolases (Other) | Lyases | Isomerases | Ligases
|