Order Center
Biochemicals
Antibiotic Explorer
BioUltra
Biological Detergents
Detection
Enzyme Explorer
BioUltra Proteins
Product Indexes
Cell Signaling Enzymes
Analytical Enzymes
Enzyme Reagents
Key Resources
Lipids
Microparticles
Peptide Explorer
Pharmacopoeia
PolyAmino Acids
BioFiles
 Enzyme Inhibitor Index
Enzyme Explorer
TO FIND AN INHIBITOR TO YOUR ENZYME...
1. Search alphabetically for the enzyme to inhibit in the table below
2. You will find the classic inhibitor and its Product Number on the same line
3. Click on the Product # to access product listing and related information including:
Product Information Sheets, C of A and MSDS
  A to C
D to K
L to O
P to Q
R to Z
A to C
Inhibited enzyme Inhibitor Product #
Acetate kinase Dimethyl dicarbonate D5520
Acetylglucosaminidase, beta-N- Acetamide A0500
Acetyl CoA-Carboxylase TOFA T6575
Acyl-CoA Cholesterol Acyltransferase NTE-122 N7911
Acyl-CoA Cholesterol Acyltransferase Sandoz 58-035 >98 % (HPLC) solid S9318
Acyl-CoA Cholesterol O-Acyltransferase YIC-C8-434 >99 % (HPLC) solid Y0628
Acyl-CoA synthase 8,11-Eicosadiynoic Acid E6517
Acyl-CoA synthase (Long Chain) 2-Fluoropalmitic Acid F4287
Adenine nucleotide translocase Bongkrekic acid solution B6179
Adenosine Kinase ABT-702 dihydrochloride A2721
Adenosine Kinase A-134974 A2846
Adenosinetriphosphatase Ouabain O3125
Adenosinetriphosphatase (microsomal calcium) Thapsigargin T9033
Adenosinetriphosphatase (Na/K) 5-(N-Ethyl-N-isopropyl)amiloride A3085
Adenosinetriphosphatase (Na/K) 5-(N-Methyl-N-isobutyl)amiloride A5585
Adenosinetriphosphatase (Na/K) Erysimoside monohydrate E5152
Adenylate cyclase 9-Cyclopentyladenine C4479
Adenylate cyclase Pyrocatechol C9510
Adenylate cyclase 2 ',5'-Dideoxyadenosine 3'-monophosphate D0689
Adenylate cyclase 2',5'-Dideoxyadenosine 3'-diphosphate D0814
Adenylate cyclase 2',5'-Dideoxyadenosine 3'-triphosphate D0939
Adenylate cyclase 2',5'-Dideoxyadenosine D7408
Adenylate cyclase 2'/3'-O-(N-Methylanthraniloyl)guanosine-5'-(g-thio)triphosphate triethylammonium salt solution >95 % (HPLC) Aqueous solution M6317
Adenylate cyclase Na-p-Tosyl-L-lysine chloromethyl ketone hydrochloride T7254
ADP ribosyltransferase 6-Nitroso-1,2-benzopyrone N8403
Akt (-)-Deguelin >98 % (HPLC) Free flowing powder D0817
Alanine aminotransferase b-Chloro-L-alanine hydrochloride C9033
Alanine hydroxymethyltransferase, D- L-Cycloserine C1159
Alanine hydroxymethyltransferase, D- DL-Cycloserine C7005
Alcohol dehydrogenase Tetraethylthiuram disulfide T1132
Alcohol dehydrogenase human 4-Methylpyrazole hydrochloride M1387
Alcohol dehydrogenase human Fomepizole M9888
Alcohol oxidase 4-Chloromercuribenzoic acid C5913
Aldehyde dehydrogenase 4-Chloromercuribenzoic acid C5913
Aldehyde dehydrogenase Tetraethylthiuram disulfide T1132
Alkylguanine-DNA alkyltransferase, mammalian DNA repair protein O6-Benzylguanine B2292
Amino acid oxidase, D- 4-Chloromercuribenzoic acid C5913
Aminopeptidase Cysteamine hydrochloride M6500
Aminopeptidase N Phebestin P7240
Aminopeptidase P Apstatin A1395
Aminotransferases O-(Carboxymethyl)hydroxylamine hemihydrochloride A4508
Amylase, alpha- a-Amylase Inhibitor from Triticum aestivum (wheat seed) A3535
Amylase, alpha- 1-Deoxynojirimycin hydrochloride D9305
Amyloglucosidase 1-Deoxynojirimycin hydrochloride D9305
Angiotensin Converting Enzyme Angiotensin Converting Enzyme Inhibitor A0773
Angiotensin Converting Enzyme Bradykinin potentiator B B0507
Angiotensin Converting Enzyme Bradykinin potentiator C B0632
Angiotensin Converting Enzyme Captopril C4042
Angiotensin Converting Enzyme Captopril C8856
Angiotensin Converting Enzyme Fenoldopam bromide F6800
Angiotensin Converting Enzyme Foroxymithine microbial F5017
Angiotensin Converting Enzyme N-Succinyl-L-proline S6633
Angiotensin Converting Enzyme Ne-Trifluoroacetyl-Lys-Pro T7025
Angiotensin Converting Enzyme Ramipril ³98% (HPLC) solid R0404
Arachidonate 12-Lipoxygenase Hydroquinone H9003
Arachidonyl synthase 8,11-Eicosadiynoic Acid E6517
Aromatase 4-Androsten-4-ol-3,17-dione A5791
Aromatase Formestane F2552
Aromatic amino acid decarboxylase, peripheral L- Benserazide hydrochloride B7283
Aromatic L-amino acid decarboxylase inhibitor S-(-)-Carbidopa C1335
Aromatic L-amino acid decarboxylase inhibitor 3-Hydroxybenzylhydrazine dihydrochloride H9382
Asparaginase 4-Chloromercuribenzoic acid C5913
Bromelain E-64 E3132
Bromelain Na-p-Tosyl-L-lysine chloromethyl ketone hydrochloride T7254
Calcineurin FK-506 powder ³98% (HPLC) F4679
Calcineurin (phosphatase activity) Calcineurin autoinhibitory fragment C3937
Calmodulin dependent Protein Kinase (CaM-KK) STO-609 >95% (HPLC), solid S1318
Calmodulin dependent Protein Kinase II Autocamtide 2-related inhibitory peptide trifluoroacetate salt A4308
Calmodulin dependent Protein Kinase II Calmodulin Kinase II Inhibitor from rat C1360
Calmodulin dependent Protein Kinase II KN-62 I2142
Calmodulin dependent Protein Kinase II KN-93 K1385
Calmodulin-regulated enzymes Calmidazolium chloride C3930
Calpain Antipain hydrochloride A6191
Calpain Acetyl-Calpastatin (184-210) Human C4285
Calpain 4-Chloromercuribenzoic acid C5913
Calpain Z-Leu-Leu-Leu-fluoromethyl keytone C8984
Calpain Z-Leu-Leu-Tyr-fluoromethyl ketone C9234
Calpain Z-Leu-Leu-Tyr-CHN2 C9859
Calpain E-64c E0514
Calpain N-Ethylmaleimide E3876
Calpain E-64d E8640
Calpain Leupeptin trifluoroacetate salt L2023
Calpain N-Methoxysuccinyl-Val-HOMO-Phe-fluoromethyl ketone M5439
Calpain a2-Macroglobulin from human plasma M6159
Calpain I N-Acetyl-Leu-Leu-Met-al A6060
Calpain I Calpain Inhibitor I A6185
Calpain I Calpain Inhibitor Peptide C9181
Calpain II N-Acetyl-Leu-Leu-Met-al A6060
Calpain II Calpain Inhibitor I A6185
Calpain II Antipain hydrochloride A6191
Calpain II Acetyl-Calpastatin (184-210) Human C4285
Calpain II 4-Chloromercuribenzoic acid C5913
Calpain II Calpain Inhibitor Peptide C9181
Calpain II E-64c E0514
Calpain II N-Ethylmaleimide E3876
Calpain II Leupeptin trifluoroacetate salt L2023
Calpain II a2-Macroglobulin from human plasma M6159
Carbonate dehydratase 1-Methylimidazole M8878
Carbonic anhydrase Acetazolamide A6011
Carbonic anhydrase Chlorothiazide C4911
Carboxylesterase Bomin-1 B6306
Carboxylesterase Bomin-2 B6431
Carboxylesterase Bomin-3 B6806
Carboxypeptidase A Carboxypeptidase Inhibitor from potato tuber C0279
Carboxypeptidase A Diethylenetriaminepentaacetic acid D6518
Carboxypeptidase A Ethylenediaminetetraacetic acid tripotassium salt dihydrate E0270
Carboxypeptidase A Ethylene glycol-bis(2-aminoethylether)-N,N,N',N'-tetraacetic acid E4378
Carboxypeptidase A 1,10-Phenanthroline monohydrate P9375
Carboxypeptidase B Carboxypeptidase Inhibitor from potato tuber C0279
Carboxypeptidase B Diethylenetriaminepentaacetic acid D6518
Carboxypeptidase B Ethylenediaminetetraacetic acid tripotassium salt dihydrate E0270
Carboxypeptidase B Ethylene glycol-bis(2-aminoethylether)-N,N,N',N'-tetraacetic acid E4378
Carboxypeptidase B 1,10-Phenanthroline monohydrate P9375
Carnitine acetyltransferase N-Ethylmaleimide E3876
Carnitine acetyltransferase Iodoacetamide I1149
Carnitine acetyltransferase Iodoacetamide I6125
Carnitine palmitoyltransferase-1 (+)-Etomoxir sodium >99 % (HPLC) powder E1905
Casein kinase 1 IC 261 ³99 % (HPLC) solid I0658
Casein Kinase G (1)-Taxifolin T4512
Casein Kinase II Myricetin M6760
Caspase Biotin-Val-Ala-Asp(OMe)-fluoromethyl ketone B2057
Caspase Boc-Asp(OMe)-fluoromethyl ketone B2682
Caspase Z-Ala-Glu(OMe)-Val-Asp(OMe)-fluoromethyl ketone C8484
Caspase Z-Val-Lys(Biotin)-Asp(OMe)-fluoromethyl ketone C8609
Caspase 1 N-Acetyl-Trp-Glu-His-Asp-al A1466
Caspase 1 Interleukin-1b (IL-1b) Converting Enzyme (ICE) Inhibitor II A4211
Caspase 1 N-Acetyl-Tyr-Val-Lys(biotinyl)-Asp 2,6-dimethylbenzoyloxymethyl ketone A4339
Caspase 1 Z-Leu-Glu(OMe)-Val-Asp(OMe)-fluoromethyl ketone C9484
Caspase 2 N-Acetyl-Val-Asp-Val-Ala-Asp-CHO A2222
Caspase 2 Z-Val-Asp(O-Me)-Val-Ala-Asp(O-Me) fluoromethyl ketone C1605
Caspase 3 N-Acetyl-Asp-Glu-Val-Asp-al A0835
Caspase 3 N-Acetyl-Ile-Glu-Thr-Asp-al A1216
Caspase 3 N-Acetyl-Glu-Ser-Met-Asp-al A1341
Caspase 3 Biotin-Asp(OMe)-Glu(OMe)-Val-Asp(OMe)-fluoromethyl ketone B1807
Caspase 3 Z-Asp(OMe)-Gln-Met-Asp(OMe) fluoromethyl ketone C0480
Caspase 3 Z-VAD-FMK V-116
Caspase 3 N-Acetyl-Leu-Glu-Val-Asp-CHO trifluoroacetate salt C0859
Caspase 5 N-Acetyl-Trp-Glu-His-Asp-al A1466
Caspase 6 N-Acetyl-Val-Glu-Ile-Asp-al A6339
Caspase 6 Biotin-Asp(OMe)-Glu(OMe)-Val-Asp(OMe)-fluoromethyl ketone B1807
Caspase 6 Z-Asp(OMe)-Gln-Met-Asp(OMe) fluoromethyl ketone C0480
Caspase 6 Z-Val-Glu(O-Me)-Ile-Asp(O-Me) fluoromethyl ketone C1730
Caspase 7 Biotin-Asp(OMe)-Glu(OMe)-Val-Asp(OMe)-fluoromethyl ketone B1807
Caspase 7 Z-Asp(O-Me)-Glu(O-Me)-Val-Asp(O-Me) fluoromethyl ketone C0605
Caspase 8 N-Acetyl-Ile-Glu-Thr-Asp-al A1216
Caspase 8 Biotin-Asp(OMe)-Glu(OMe)-Val-Asp(OMe)-fluoromethyl ketone B1807
Caspase 8 Z-Ile-Glu(O-ME)-Thr-Asp(O-Me) fluoromethyl ketone C1230
Caspase 8 Z-Leu-Glu(OMe)-Thr-Asp(OMe)-fluoromethyl ketone C8734
Caspase 9 Z-Leu-Glu(O-ME)-His-Asp(O-Me) fluoromethyl ketone C1355
Caspase 10 Biotin-Asp(OMe)-Glu(OMe)-Val-Asp(OMe)-fluoromethyl ketone B1807
Caspase 13 Z-Leu-Glu(OMe)-Glu(OMe)-Asp(OMe)-fluoromethyl ketone C8859
Catalase 3-Amino-1,2,4-triazole A8056
Cathepsin B N-Acetyl-Leu-Leu-Met-al A6060
Cathepsin B Calpain Inhibitor I A6185
Cathepsin B Antipain hydrochloride A6191
Cathepsin B Biotin-Phe-Ala-fluoromethyl ketone B1932
Cathepsin B Cystatin from chicken egg white C0408
Cathepsin B CA-074 C5732
Cathepsin B CA-074 methyl ester C5857
Cathepsin B Chymostatin C7268
Cathepsin B Cystatin from chicken egg white C8917
Cathepsin B Z-Phe-Phe-fluoromethyl keytone C9109
Cathepsin B E-64 E3132
Cathepsin B Leupeptin trifluoroacetate salt L2023
Cathepsin B N-Methoxysuccinyl-Phe-HOMO-Phe-fluoromethyl ketone M5314
Cathepsin B a2-Macroglobulin from human plasma M6159
Cathepsin B Procathepsin B Fragment 26-50, rat P0350
Cathepsin B Procathepsin B Fragment 36-50, rat P0475
Cathepsin D Antipain hydrochloride A6191
Cathepsin D Chymostatin C7268
Cathepsin D Pepstatin A P5318
Cathepsin G a1-Antichymotrypsin from human plasma A9285
Cathepsin G Chymostatin C7268
Cathepsin G Z-Gly-Leu-Phe-chloromethyl ketone C9984
Cathepsin G Diisopropyl fluorophosphate D0879
Cathepsin G 3,4-Dichloroisocoumarin D7910
Cathepsin G N-Acetyl-eglin C E7888
Cathepsin L Calpain Inhibitor I A6185
Cathepsin L Z-Phe-Phe-fluoromethyl keytone C9109
Cathepsin L Z-Leu-Leu-Tyr-fluoromethyl ketone C9234
Cathepsin L Z-Leu-Leu-Tyr-CHN2 C9859
Cathepsin L N-Methoxysuccinyl-Phe-HOMO-Phe-fluoromethyl ketone M5314
Cdc2-like Kinase TG003 T5575
Ceramidase N-Oleoylethanolamine O0383
Chloroperoxidase 1-Aminobenzotriazole A3940
Cholesterol esterase Diethylumbelliferyl phosphate oil >99% (HPLC) D7692
Cholesterol-O-Acetyltransferase NTE-122 Dihydrochloride solid, ≥97% (HPLC) N7911
Choline acetyltransferase 2-(a-Naphthoyl)ethyltrimethylammonium iodide N7906
Choline acetyltransferase Acetylethylcholine mustard hydrochloride C-001
Choline dehydrogenase Betaine aldehyde chloride B3650
Cholinesterase Deoxypeganine hydrochloride dihydrate D7314
Cholinesterase Galanthamine hydrobromide from Lycoris sp. G1660
Cholinesterase Hydroxytacrine maleate salt H3146
Cholinesterase, acetyl 9-Amino-1,2,3,4-tetrahydroacridine hydrochloride A3773
Cholinesterase, acetyl 1,5-Bis(4-allyldimethylammoniumphenyl)pentan-3-one dibromide A9013
Cholinesterase, acetyl 3,4-Dichloroisocoumarin D7910
Cholinesterase, acetyl Edrophonium chloride E3256
Cholinesterase, acetyl Fisonate F6051
Cholinesterase, acetyl (1)-Huperzine A H5777
Cholinesterase, acetyl Peganole P3240
Cholinesterase, acetyl Pyridostigmine bromide P9797
Cholinesterase, butyryl Betaine anhydrous B2629
Cholinesterase, butyryl Betaine inner salt monohydrate B2754
Cholinesterase, butyryl Betaine hydrochloride B3501
Cholinesterase, butyryl Pyridostigmine bromide P9797
Cholinesterase, butyryl Tetraisopropyl pyrophosphoramide T1505
Chymotrypsin A gamma, Bovine Z-L-Phe chloromethyl ketone C9511
Chymotrypsin, alpha- a2-Antiplasmin from human plasma A0914
Chymotrypsin, alpha- Aprotinin from bovine lung A1153
Chymotrypsin, alpha- 6-Aminohexanoic acid A2504
Chymotrypsin, alpha- a1-Antitrypsin from human plasma A6150
Chymotrypsin, alpha- 6-Aminohexanoic acid A7824
Chymotrypsin, alpha- 4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride A8456
Chymotrypsin, alpha- a1-Antitrypsin from human plasma A9024
Chymotrypsin, alpha- a1-Antichymotrypsin from human plasma A9285
Chymotrypsin, alpha- Bromoenol lactone B1552
Chymotrypsin, alpha- Chymostatin C7268
Chymotrypsin, alpha- Diisopropyl fluorophosphate D0879
Chymotrypsin, alpha- Ecotin E1398
Chymotrypsin, alpha- N-Acetyl-eglin C E7888
Chymotrypsin, alpha- Gabexate mesylate G2417
Chymotrypsin, alpha- Leupeptin trifluoroacetate salt L2023
Chymotrypsin, alpha- Trypsin inhibitor from bovine pancreas T0256
Chymotrypsin, alpha- Trypsin inhibitor from chicken egg white T1886
Chymotrypsin, alpha- N-p-Tosyl-L-phenylalanine chloromethyl ketone T4376
Chymotrypsin, alpha- Trypsin inhibitor from turkey egg white T4385
Chymotrypsin, alpha- Trypsin inhibitor from chicken egg white T9253
Chymotrypsin, alpha- Trypsin inhibitor from Phaseolus limensis (lima bean) T9378
Chymotrypsin, alpha- Trypsin-chymotrypsin inhibitor from Glycine max (soybean) T9777
Citrate Lyase MEDICA-16 M5693
Clostripain 4-Chloromercuribenzoic acid C5913
Clostripain Histatin 5 H6027
Clostripain Na-p-Tosyl-L-lysine chloromethyl ketone hydrochloride T7254
Creatine Phosphokinase 4-Chloromercuribenzoic acid C5913
Collagenase 5-(R,S)-T-trans-cinnamido-7-methyl-4-oxo-octanoyl-Pro-Pro C0360
Collagenase Dichloromethylenediphosphonic acid disodium salt D4434
Collagenase Ecotin E1398
Collagenase Isoamylphosphonyl-Gly-Pro-Ala dipotassium salt I7029
Collagenase (2R)-2-Mercaptomethyl-4-methylpentanoyl-b-(2-naphthyl)-Ala-Ala Amide M3531
Fibroblast collagenase N-Phospho-Ile(O-ethyl)-Tyr(O-benzyl)-Gly dipotassium salt P6115
Complement C1 Esterase inhibitor C-1 from human plasma E0518
Complement C1r Esterase inhibitor C-1 from human plasma C2412
Complement C1r Diisopropyl fluorophosphate D0879
Complement C1r Esterase inhibitor C-1 from human plasma E0518
Complement C1s Esterase inhibitor C-1 from human plasma C2412
Complement C1s Diisopropyl fluorophosphate D0879
Complement C1s Esterase inhibitor C-1 from human plasma E0518
Complement Factor B Diisopropyl fluorophosphate D0879
Complement Factor D Diisopropyl fluorophosphate D0879
Cyclin Dependant Kinase Compound 52 C2604
Cyclin Dependant Kinase Indirubin-3'-oxime I0404
Cyclin Dependant Kinase N9-Isopropylolomoucine I0763
Cyclin-Dependent Kinase Olomoucine O0886
Cyclin-dependent Kinase [Ala92]-Peptide 6 P7967
Cyclin-dependent kinases Roscovitine R7772
Cyclin Dependant Kinase Roscovitine R7772
Cyclin Dependant Kinase I Butyrolactone I B7930
Cyclin Dependant Kinase I Alsterpaullone A4847
Cyclin Dependant Kinase 2 GW8510 G7791
Cyclin Dependant Kinase 2 NU2058 >98 % (HPLC) N4286
Cyclin Dependant Kinase 2