Order Center
Biochemicals
Antibiotic Explorer
BioUltra
Biomaterials
Detection
Detergent Theory
Enzyme Explorer
BioUltra Proteins
Product Indexes
Cell Signaling Enzymes
Analytical Enzymes
Enzyme Reagents
Key Resources
Lipids
Microparticles
Pharmacopoeia
PolyAmino Acids
Reference Chart
BioFiles
 Enzyme Inhibitor Index
Enzyme Explorer
TO FIND AN INHIBITOR TO YOUR ENZYME...
1. Search alphabetically for the enzyme to inhibit in the table below
2. You will find the classic inhibitor and its Product Number on the same line
3. Click on the Product # to access product listing and related information including:
Product Information Sheets, C of A and MSDS
  A to C
D to K
L to O
P to Q
R to Z
D to K
Inhibited enzyme Inhibitor Product #
7-Dehydrocholesterol reductase BM 15766 sulfate powder ³99% (HPLC) B8685
Desmosteroldelta 24 reductase Triparanol T5200
Deoxyribonuclease I Ethylenediaminetetraacetic acid tripotassium salt dihydrate E0270
Deoxyribonuclease I Ethylene glycol-bis(2-aminoethylether)-N,N,N',N'-tetraacetic acid E4378
Deoxyribonuclease I Netropsin dihydrochloride from Streptomyces netropsis N9653
Deoxyribonuclease I 1,10-Phenanthroline monohydrate P9375
D5-Desaturase Sesamin S9314
D5/D6-Desaturase CP-24879 hydrochloride solid >98% C9115
Diacylglycerol kinase Diacylglycerol Kinase Inhibitor II D5794
Diacylglycerol kinase Diacylglycerol Kinase Inhibitor I D5919
Diacylglycerol kinase 1,2-Dioctanoylethylene glycol D8907
Diacylglycerol kinase RHC 80267 ³98 % (HPLC) R2028
Diaphorase Carmustine C0400
Dihydrofolate reductase Aminopterin A1784
Dihydrofolate reductase Methotrexate hydrate A6770
Dihydrofolate reductase (1)Amethopterin A7019
Dihydrofolate reductase 4-Chloromercuribenzoic acid C5913
Dihydroorotate dehydrogenase Leflunomide L5025
Dipeptidyl peptidase IV Ile-Pro-Ile I9759
DNA polymerase - α Cephalomannine ≥97% (HPLC), solid C4991
DNA polymerase and proline dipeptidase Phosphonoacetic acid P6909
Dopamine beta-hydroxylase 1-(4-Hydroxybenzyl)imidazole-2-thiol H6892
Dopamine beta-hydroxylase 1-Phenyl-3-(2-thiazolyl)-2-thiourea P4015
Elastase, granulocyte N-(Methoxysuccinyl)-Ala-Ala-Pro-Val-chloromethyl ketone M0398
Elastase, leukocyte Aprotinin from bovine lung A1153
Elastase, leukocyte Diisopropyl fluorophosphate D0879
Elastase, leukocyte 3,4-Dichloroisocoumarin D7910
Elastase, leukocyte Eglin C fragment 60-63 methyl ester E2017
Elastase, leukocyte N-Acetyl-eglin C E7888
Elastase, leukocyte Elafin Human E7280
Elastase, leukocyte a2-Macroglobulin from human plasma M6159
Elastase, leukocyte Phenylmethanesulfonyl fluoride P7626
Elastase, neutrophil GW311616A G8419
Elastase, pancreatic a1-Antitrypsin from human plasma A6150
Elastase, pancreatic a1-Antitrypsin from human plasma A9024
Elastase, pancreatic Diisopropyl fluorophosphate D0879
Elastase, pancreatic 3,4-Dichloroisocoumarin D7910
Elastase, pancreatic Ecotin E1398
Elastase, pancreatic a2-Macroglobulin from human plasma M6159
Elongation factor-2 Kinase NH125 solid ³98% (HPLC) N2162
Endothelin converting enzyme SM-19712 ³98% (HPLC) solid S3445
Enkephalinase DL-Thiorphan T6031
Esterase, lipase and N-formylmethionine aminopeptidase Ebelactone A microbial E0761
Esterase, lipase and N-formylmethionine aminopeptidase. Ebelactone B microbial E0886
Estrone sulfatase Estrone sulfamate E9645
Factor VIIIa Acetyl-Ala-Leu-Cys-Asp-Asp-Pro-Arg-Val-Asp-Arg-Trp-Tyr-Cys-Gln-Phe-Val-Glu-Gly-NH2 [Disulfide bridge: 3-13] trifluoroacetate salt A0472
Factor Xa a2-Antiplasmin from human plasma A0914
Factor Xa Antithrombin III from rat plasma A6397
Factor Xa Antithrombin III from human plasma A2221
Factor Xa Antithrombin III from bovine plasma A9141
Factor Xa Diisopropyl fluorophosphate D0879
Factor Xa 3,4-Dichloroisocoumarin D7910
Factor Xa Ecotin E1398
Factor XIIa Esterase inhibitor C-1 from human plasma E0518
Farnesyl diphosphate synthase Etidronate disodium solid P5248
Farnesyl diphosphate synthase Pamidronate disodium >99 % (HPLC) powder P2371
Farnesyl transferase a-Hydroxyfarnesylphosphonic acid H9279
Farnesyl transferase FTI-276 F9553
Farnesyl transferase FTI-277 F9803
Farnesyl transferase, p21ras Cys-Val-2-Nal-Met C4433
Farnesyl transferase, Ras Cys-4-(2-aminobenzoyl)-Met C5684
Farnesyl transferase, Ras L-744,832 L7287
Farnesyl transferase, Ras Manumycin A from Streptomyces parvulus M6418
Fatty acid synthase Cerulenin from Cephalosporium caerulens C2389
Fatty acid synthase 4-Methylene-2-octyl-5-oxotetrahydrofuran-3-carboxylic acid C5490
Fatty acid amide hydrolase Oleyl trifluoromethyl ketone O9887
Ficin Cystatin from chicken egg white C8917
Ficin E-64 E3132
Follicle-stimulating hormone-suppressing protein Inhibin from porcine ovary I9149
Fusarinine -C Ornithinesterase N-Bromoacetamide B2377
Fusarinine -C Ornithinesterase DL-Buthionine-[S,R]-sulfoximine B2640
Galactose oxidase Iodoacetamide I1149
Galactose oxidase Iodoacetamide I6125
Galactosidase, alpha- Deoxygalactonojirimycin hydrochloride D9641
Galactosidase, beta- Diethylenetriaminepentaacetic acid D6518
Galactosidase, beta- Ethylenediaminetetraacetic acid tripotassium salt dihydrate E0270
Galactosidase, beta- Ethylene glycol-bis(2-aminoethylether)-N,N,N',N'-tetraacetic acid E4378
Galactosidase, beta- Ethylenediamine E4379
Galactosidase, beta- Nojirimycin bisulfite microbial N7779
Galactosidase, beta- 1,10-Phenanthroline monohydrate P9375
Galactosidase, beta- Quercetin dihydrate Q0125
Geranylgeranyl transferase I GGTI-2133 G5294
Geranylgeranyl transferase I GGTI-298 G5169
Glucanase, exo-beta-(1,3)- 2,4-Dinitrophenyl 2-fluoro-2-deoxy-b-D-glucopyranoside D1791
Glucosidase 6-Bromo-4-cyclohexene-1,2,3-triol Mixed Isomers B1147
Glucosidase I N-Methyl-1-deoxynojirimycin M1777
Glucosidase II (not glucosidase I) 6-Bromo-4-cyclohexene-1,2,3-triol Mixed Isomers B1147
Glucosidase, alpha- N-Butyldeoxynojirimycin B8299
Glucosidase, alpha- Castanospermine from Castanospermum australe seeds C3784
Glucosidase, alpha- 1,4-Dideoxy-1,4-imino-D-arabinitol D1542
Glucosidase, alpha-, yeast 6-Bromo-4-cyclohexene-1,2,3-triol Mixed Isomers B1147
Glucosidase, beta- Conduritol B epoxide C5424
Glucosidase, beta- 4-Chloromercuribenzoic acid C5913
Glucosidase, beta- 1-Deoxynojirimycin hydrochloride D9305
Glucosylceramide synthase inhibitor (1)-threo-1-Phenyl-2-decanoylamino-3-morpholino-1-propanol hydrochloride P7340
beta-Glucuronidase D-Glucaro-d-lactam potassium salt G7166
Glutamate carboxypeptidase II PBDA P2238
Glutamic Decarboxylase, L- Chelidamic aicd C8011
Glutamic Decarboxylase, L- Dimethyl dicarbonate D5520
Glutamyltranspeptidase, gamma- Sodium iodoacetate I2512
Glutamyltranspeptidase, gamma- Iodoacetic acid I4386
Glutamyltranspeptidase, gamma- Sodium iodoacetate I9148
Glutathione biosynthesis and Glu-Cys Ligase L-Buthionine-sulfoximine B2515
Glutathione Reductase Carmustine C0400
Glutathione S-transferase S-(p-Azidophenacyl)glutathione A1782
Glutathione S-transferase Ebselen E3520
Glutathione S-transferase (1)-Taxifolin T4512
Glutathione S-transferase Sodium taurocholate T9034
Glyceraldehyde 3-Phosphate Dehydrogenase from a green algae N,N,N',N'-Tetramethylazodicarboxamide D3648
Glyceraldehyde-3-phosphate dehydrogenase Sodium iodoacetate I2512
Glyceraldehyde-3-phosphate dehydrogenase Iodoacetic acid I4386
Glyceraldehyde-3-phosphate dehydrogenase Sodium iodoacetate I9148
Glycogen synthase kinase TDZD-8 needles >99% T8325
Glycosidase inhibitor (2R,5R)-Bis(hydroxymethyl)-(3R,4R)-dihydroxypyrrolidine H9632
Glycosyltransferase, Ceramide-specific N-Butyldeoxynojirimycin B8299
Glyoxalase I S-(p-Azidophenacyl)glutathione A1782
Granzyme B Antipain hydrochloride A6191
Granzyme B Z-Ala-Ala-Asp(OMe)-chloromethyl ketone C0235
Granzyme B 3,4-Dichloroisocoumarin D7910
Guanylate cyclase 4-Phenyl-3-furoxancarbonitrile P1726
Guanylyl cyclase 1H-[1,2,4]Oxadiazolo[4,3-a]quinoxalin-1-one O3636
Hexokinase Quercetin dihydrate Q0125
Hexokinase (mitochondrial), cellular respiration, and glycolysis Lonidamine L4900
Histidine ammonia-lyase and decarboxylase 4-Imidazoleacrylic acid U7500
Histidine kinase NH125 N2162
Histidinol dehydrogenase 1-Methylimidazole M8878
Histinol Dehydrogenase 4-Imidazolemethanol hydrochloride H1877
Histone deacetylase APHA Compound 8 ³99 % (HPLC) solid A2478
Histone deacetylase Apicidin ³95 % from microbial source solid A8851
Histone deacetylase (-)-Depudecin >95 % (HPLC) from microbial source D5816
Histone deacetylase Scriptaid S7817
Histone deacetylase Sirtinol ~95 % S7942
Histone deacetylase Sodium butyrate ~99 % B5887
Histone deacetylase Trichostatin A from Streptomyces sp. T8552
HIV integrase His-Cys-Lys-Phe-Trp-Trp H6387
HIV Protease a-MAPI M9812
Hydrolases or "isopeptidases." Ubiquitin Aldehyde U1507
Hydroxymethylglutaryl-CoA Reductase Mevastatin M2537
Hydroxymethylglutaryl-CoA Reductase Mevinolin from Aspergillus sp. M2147
Hydroxymethylglutaryl-CoA Reductase Pravastatin sodium powder >98% (HPLC) P4498
Hydroxymethylglutaryl-CoA Reductase SR 12813 S4194
15-Hydroxyprostaglandin Dehydrogenase CAY10397 C6734
IKK-2 IMD-0354 I3159
Inosine monophosphate (IMP) dehydrogenase Mycophenolic acid from Penicillium brevi-compactum M3536
Inosine monophosphate (IMP) dehydrogenase Ribavirin R9644
Integrase (-)-Arctigenin A1854
Isocitric dehydrogenase (NADP) Diethylenetriaminepentaacetic acid D6518
Isocitric dehydrogenase (NADP) Ethylenediaminetetraacetic acid tripotassium salt dihydrate E0270
Isocitric dehydrogenase (NADP) Ethylene glycol-bis(2-aminoethylether)-N,N,N',N'-tetraacetic acid E4378
Isocitric dehydrogenase (NADP) 1,10-Phenanthroline monohydrate P9375
Isocitric dehydrogenase (NADP) 1,10-Phenanthroline monohydrate P9375
Jak-2 protein tyrosine kinase (PTK) Tyrphostin AG 490 T3434
Jak-3 protein tyrosine kinase (PTK) ZM 39923 Z4626
c-Jun N-terminal kinase JIP-1 (153-163) ³98 % (HPLC) Lyophilized powder J2270
Kallikrein tissue a2-Antiplasmin from human plasma A0914
Kallikrein tissue Aprotinin from bovine lung A1153
Kallikrein tissue Antithrombin III from human plasma A2221
Kallikrein tissue Antithrombin III from bovine plasma A9141
Kallikrein tissue Benzamidine hydrochloride hydrate B6506
Kallikrein tissue Diisopropyl fluorophosphate D0879
Kallikrein tissue 3,4-Dichloroisocoumarin D7910
Kallikrein tissue Leupeptin trifluoroacetate salt L2023
Kallikrein, Plasma Esterase inhibitor C-1 from human plasma E0518
Kallikrein, Plasma Gabexate mesylate G2417
  back to top
A to C D to K L to O P to Q R to Z