Sigma-Aldrich
Sigma - Life Science Logo
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Enzyme Explorer
L to O
TO FIND AN INHIBITOR TO YOUR ENZYME...
1. Search alphabetically for the enzyme to inhibit in the table below
2. You will find the classic inhibitor and its Product Number on the same line
3. Click on the Product # to access product listing and related information including:
Product Information Sheets, C of A and MSDS
  A to C
D to K
L to O
P to Q
R to Z
L to O
Inhibited enzyme Inhibitor Product #
Lactic Dehydrogenase, Bovine (H4 isozyme) 2,3-Butanedione D3634
Lactic dehydrogenase, L- Sodium oxamate O2751
Lck 7-Cyclopentyl-5-(4-phenoxyphenyl)-7H-pyrrolo[2,3‑d]pyrimidin-4-ylamine ³99 % (HPLC) C8863
Leucine Aminopeptidase (2S,3R)-3-Amino-2-hydroxy-4-(4-nitrophenyl)butanoyl-L-leucine hydrochloride A5921
Leucine Aminopeptidase Actinonin A6671
Leucine Aminopeptidase Bestatin hydrochloride B8385
Leucine aminopeptidase, cytosol Amastatin hydrochloride A1276
Leucine aminopeptidase, cytosol (2S,3R)-3-Amino-2-hydroxy-4-(4-nitrophenyl)butanoyl-L-leucine hydrochloride A5921
Leucine aminopeptidase, cytosol Actinonin A6671
Leucine aminopeptidase, cytosol Bestatin hydrochloride B8385
Leucine aminopeptidase, cytosol Diethylenetriaminepentaacetic acid D6518
Leucine aminopeptidase, cytosol 3,4-Dichloroisocoumarin D7910
Leucine aminopeptidase, cytosol Ethylenediaminetetraacetic acid tripotassium salt dihydrate E0270
Leucine aminopeptidase, cytosol Ethylene glycol-bis(2-aminoethylether)-N,N,N',N'-tetraacetic acid E4378
Leucine aminopeptidase, cytosol 1,10-Phenanthroline monohydrate P9375
Leucine aminopeptidase, microsomal Amastatin hydrochloride A1276
Leucine aminopeptidase, microsomal (2S,3R)-3-Amino-2-hydroxy-4-(4-nitrophenyl)butanoyl-L-leucine hydrochloride A5921
Leucine aminopeptidase, microsomal Actinonin A6671
Leucine aminopeptidase, microsomal Bestatin hydrochloride B8385
Leukotriene A4 hydrolase SC 22716 >99 % (HPLC) P7495
Lipase, Pancreatic Orlistat O4139
Lipoxygenase 7,7-Dimethyl-(5Z,8Z)-eicosadienoic acid D8008
Lipoxygenase 5,8,11,14-Eicosatetraynoic acid E1768
Lipoxygenase Nordihydroguaiaretic acid from Larrea divaricata (creosote bush) N5023
Lipoxygenase 9,12-Octadecadiynoic acid O5636
Lipoxygenase (5 and 12-); does not inhibit cyclooxygenase. 5,8,11-Eicosatriynoic acid E3140
Lipoxygenase (5-) AA-861 A3711
Lipoxygenase (5-) BW A4C ³98 % (HPLC) B7559
Lipoxygenase (5-) 5,6-Dehydroarachidonic acid D2154
Lipoxygenase (5-) cis-5,8,11,14,17-Eicosapentaenoic acid E2011
Lipoxygenase (5-) cis-5,8,11,14,17-Eicosapentaenoic acid sodium salt E6627
Lipoxygenase (5-) 15S-Hydroxy-5Z,8Z,11Z,13E-eicosatetraene-(2-biotinyl)hydrazide H6785
Lipoxygenase (5-) Piriprost potassium salt P6113
Lipoxygenase (5-) REV 5901 R5523
Lipoxygenase (5-) and cyclooxygenase Meclofenamic acid sodium salt M4531
Lipoxygenase (15-) PD 146176 >99 % (HPLC) solid P4620
Lysyl oxidase 3-Aminopropionitrile fumarate salt A3134
Mannosidase, alpha- 4-Chloromercuribenzoic acid C5913
Mannosidase, alpha- Diethylenetriaminepentaacetic acid D6518
Mannosidase, alpha- 1,4-Dideoxy-1,4-imino-D-mannitol hydrochloride D8390
Mannosidase, alpha- Ethylenediaminetetraacetic acid tripotassium salt dihydrate E0270
Mannosidase, alpha- Ethylene glycol-bis(2-aminoethylether)-N,N,N',N'-tetraacetic acid E4378
Mannosidase, alpha- 1,10-Phenanthroline monohydrate P9375
Mannosidase, alpha- Swainsonine S6640
Mannosidase, alpha- Swainsonine S9263
MAP Kinase Apigenin A3145
MAP Kinase SB 203770 S8307
MAP Kinase II CL-100/HVH1/Mkp1 human C7984
MAPKAP kinase-1beta and p70 S6 kinase Ro 32-0432 hydrochloride R-137
MAPKAP kinase-1beta and p70 S6 kinase Ro 31-8220 methanesulfonate salt R-136
MAPKAP kinase-1beta and p70 S6 kinase Ro 32-0432 hydrochloride R-137
Matrix metalloprotease N-[(2S)-(Methoxycarbonylmethyl)-4-methylpentanoyl]-L-tryptophan-methylamide M5939
Metalloprotease inhibitor 2,2'-Bipyridyl D7505
Metallo-protease inhibitor with selectivity for aminopeptidases. Epiamastatin hydrochloride E3389
Metallo-protease inhibitor, aminopeptidases M and B L-Leucinethiol, oxidized dihydrochloride L8397
Metallo-protease selectively aminopeptidases Epibestatin hydrochloride E0381
Methionine Aminopeptidase-2 Fumagillin F6771
Monoamine oxidase competitive inhibitor Debrisoquine sulfate salt D1306
Monoamine oxidase inhibitor R-(-)-Deprenyl hydrochloride M-003
Monoamine oxidase inhibitor S(+)-Deprenyl hydrochloride M-005
Monoamine oxidase inhibitor Hydroxylamine hydrochloride H9876
Monoamine oxidase inhibitor Hydralazine hydrochloride H1753
Monoamine oxidase inhibitor Hydroxylamine sulfate salt H0127
Monoamine oxidase inhibitor Iproniazid phosphate salt I7627
Monoamine oxidase inhibitor Nialamide N1392
Monoamine oxidase inhibitor Pargyline hydrochloride P8013
Monoamine oxidase inhibitor Phenelzine sulfate salt P6777
Monoamine oxidase inhibitor trans-2-Phenylcyclopropylamine hemisulfate salt P1761
Monoamine oxidase inhibitor trans-2-Phenylcyclopropylamine hydrochloride P8511
Monoamine oxidase inhibitor Quinacrine dihydrochloride Q3251
Monoamine oxidase inhibitor Ro 16-6491 hydrochloride R-106
Monoamine oxidase inhibitor Ro 41-1049 hydrochloride R-107
Monoamine Oxidase-A N-Methyl-N-propargyl-3-(2,4-dichlorophenoxy)propylamine hydrochloride M3778
MRP 1-Benzoyl-5-methoxy-2-methylindole-3-acetic acid B1183
MRP 1-Benzoyl-5-methoxy-2-methylindole-3-acetic acid B1183
MRP 1-(4-Chlorobenzyl)-5-methoxy-2-methylindole-3-acetic acid C1610
MRP 1-(4-Fluorobenzyl)-5-methoxy-2-methylindole-3-acetic acid F2927
Myelin Basic Protein Kinase Ala-Pro-Arg-Thr-Pro-Gly-Gly-Arg-Arg A6583
Myosin Light Chain Kinase HA-100 I1392
Myosin Light Chain Kinase ML-7 I2764
Myosin Light Chain Kinase ML-9 C1172
NADH peroxidase Diethylenetriaminepentaacetic acid D6518
NADH peroxidase Ethylenediaminetetraacetic acid tripotassium salt dihydrate E0270
NADH peroxidase Ethylene glycol-bis(2-aminoethylether)-N,N,N',N'-tetraacetic acid E4378
NADH peroxidase 1,10-Phenanthroline monohydrate P9375
Nitric Oxide Synthase S-(2-Aminoethyl)isothiouronium bromide hydrobromide A5879
Neuraminidase Siastatin B microbial S8063
Nitric Oxide Synthase Aminoguanidine hemisulfate A7009
Nitric Oxide Synthase Aminoguanidine bicarbonate salt A7259
Nitric Oxide Synthase Aminoguanidine hydrochloride A8835
Nitric Oxide Synthase Arcaine sulfate salt A0384
Nitric Oxide Synthase Carboxy-PTIO potassium salt C-221
Nitric Oxide Synthase Chlorpromazine hydrochloride C8138
Nitric Oxide Synthase Diphenyleneiodonium chloride D2926
Nitric Oxide Synthase NG,NG-Dimethylarginine hydrochloride D4268
Nitric Oxide Synthase S-Ethylisothiourea hydrobromide E3149
Nitric Oxide Synthase a-Guanidinoglutaric acid G7788
Nitric Oxide Synthase NG-Hydroxy-L-arginine acetate salt H7278
Nitric Oxide Synthase 2-Iminobiotin I4632
Nitric Oxide Synthase L-N6-(1-Iminoethyl)lysine hydrochloride I8021
Nitric Oxide Synthase L-N5-(1-Iminoethyl)ornithine hydrochloride I8768
Nitric Oxide Synthase Melatonin M5250
Nitric Oxide Synthase Nw-Nitro-L-arginine N5501
Nitric Oxide Synthase Nw-Nitro-L-arginine methyl ester hydrochloride N5751
Nitric Oxide Synthase L-Nϖ-Nitroarginine-2,4-L-diaminobutyric amide di(trifluoroacetate) salt ³95 % (HPLC) powder N3411
Nitric Oxide Synthase N-[(4S)-4-amino-5-[(2-aminoethyl)amino]pentyl]-N'-nitroguanidine tris(trifluoroacetate) salt ³98 % (HPLC) powder A5727
Nitric Oxide Synthase 7-Nitroindazole N7778
Nitric Oxide Synthase 1,3-PBIT dihydrobromide P8227
Nitric Oxide Synthase 1,4-PBIT dihydrobromide P8352
Nitric Oxide Synthase TFPI hydrochloride T7188
Nitric Oxide Synthase 1-[2-(Trifluoromethyl)phenyl]imidazole T7313
Nitric Oxide Synthase (iNOS) 2-Amino-5,6-dihydro-6-methyl-4H-1,3-thiazine A9834
Nitric Oxide Synthase (iNOS) 3-Bromo-7-nitroindazole B2050
Nitric Oxide Synthase (iNOS) L-Canavanine from Canavalia ensiformis (Jack bean) C1625
Nitric Oxide Synthase (iNOS) L-Canavanine sulfate C9758
Nitric Oxide Synthase (iNOS) Curcumin C1386
Nitric Oxide Synthase (iNOS) Guanidinoethyl disulfide dihydrobromide G4788
Nitric Oxide Synthase (iNOS) L-N6-(1-Iminoethyl)lysine hydrochloride I8021
Nitric Oxide Synthase (iNOS) Mercaptoethylguanidine sodium succinate M-276
Nitric Oxide Synthase (iNOS) S-Methylisothiourea hemisulfate M3127
Nitric Oxide Synthase (iNOS) NG-Methyl-L-arginine acetate salt M7033
Nitric Oxide Synthase (iNOS) 1,3-PBIT dihydrobromide P8227
Nitric Oxide Synthase (iNOS) 1,4-PBIT dihydrobromide P8352
Nitric Oxide Synthase neuronal 3-Bromo-7-nitroindazole B2050
Nitric Oxide Synthase neuronal Proadifen hydrochloride P1061
Nitric Oxide Synthase neuronal Spermidine S2626
Nitric Oxide Synthase neuronal Spermine S3256
Nucleotidase, 5'- Diethylenetriaminepentaacetic acid D6518
Nucleotidase, 5'- Ethylenediaminetetraacetic acid tripotassium salt dihydrate E0270
Nucleotidase, 5'- Ethylene glycol-bis(2-aminoethylether)-N,N,N',N'-tetraacetic acid E4378
Nucleotidase, 5'- 1,10-Phenanthroline monohydrate P9375
back to top
A to C D to K L to O P to Q R to Z