| R to Z |
| Inhibited enzyme |
Inhibitor |
Product # |
| Renin |
Pepstatin A |
P5318 |
| Renin, human |
D-His-Pro-Phe-His-Leu-y-(CH2NH)-Leu-Val-Tyr |
H6137 |
| Reverse Transcriptase, AMV |
Nalidixic acid |
N8878 |
| Rho-Associated protein kinase |
Y-27632 dihydrochloride >99 % (HPLC) solid |
Y0503 |
| Rho-Kinase |
Hydroxyfasudil solid ³98 % (HPLC) |
H4413 |
| Rho-Kinase |
Y-27632 dihydrochloride >99 % (HPLC) solid |
Y0503 |
| Ribonuclease A |
Diethyl pyrocarbonate |
D5758 |
| Ribonuclease A |
Ribonuclease Inhibitor from human placenta |
R4380 |
| Ribonuclease A |
Ribonuclease Inhibitor from human placenta |
R7253 |
| RNA Polymerase |
Thiolutin from Streptomyces luteosporeus |
T3450 |
| APP-Secretase |
[Asn670, Sta671, Val672]-Amyloid b/A4 Precursor Protein 770 Fragment 662-675 trifluoroacetate salt |
A1847 |
| β-Secretase |
Glu-Val-Asn-[(2R,4S,5S)-5-amino-4-hydroxy-2,7-dimethyloctanoyl]-Ala-Glu-Phe trifluoroacetate salt |
G8291 |
| β-Secretase |
b-Secretase Inhibitor |
S4562 |
| γ Secretase |
[(2R,4R,5S)-2-Benzyl-5-(Boc-amino)-4-hydroxy-6-phenylhexanoyl]-Leu-Phe-NH2 trifluoroacetate salt |
B5306 |
| γ Secretase |
DAPT |
D5942 |
| γ Secretase |
g-Secretase Inhibitor |
S2188 |
| Spermidine synthase |
Cysteamine hydrochloride |
M6500 |
| Spermine synthase |
Cysteamine hydrochloride |
M6500 |
| Sphingomyelinase (neutral) |
GW4869 solid ~91 % (HPLC) |
D1692 |
| Sphingosine acyltransferase |
Fumonisin B2 from Fusarium moniliforme |
F3771 |
| Sphingosine N-acyltransterase |
Fumonisin B1 from Fusarium moniliforme |
F1147 |
| Squalene epoxidase |
NB-598 solid ~97 % |
N4536 |
| Squalene Synthase |
YM-53601 solid >98 % (HPLC) |
Y0128 |
| Src Kinase |
SU 6656 >99 % (HPLC) powder |
S9692 |
| Succinyl Dehydrogenase |
3-Nitropropionic acid |
N5636 |
| Syk |
ER-27319 |
E0656 |
| Thermolysin |
Diethylenetriaminepentaacetic acid |
D6518 |
| Thermolysin |
Ethylenediaminetetraacetic acid tripotassium salt dihydrate |
E0270 |
| Thermolysin |
Ethylene glycol-bis(2-aminoethylether)-N,N,N',N'-tetraacetic acid |
E4378 |
| Thermolysin |
a2-Macroglobulin from human plasma |
M6159 |
| Thermolysin |
1,10-Phenanthroline monohydrate |
P9375 |
| Thermolysin |
Phosphoramidon disodium salt |
R7385 |
| Thrombin |
a2-Antiplasmin from human plasma |
A0914 |
| Thrombin |
Antipain hydrochloride |
A6191 |
| Thrombin |
Antithrombin III from human plasma |
A2221 |
| Thrombin |
Antithrombin III from rat plasma |
A6397 |
| Thrombin |
Antithrombin III from human plasma |
A2221 |
| Thrombin |
4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride |
A8456 |
| Thrombin |
Antithrombin III from bovine plasma |
A9141 |
| Thrombin |
Diisopropyl fluorophosphate |
D0879 |
| Thrombin |
3,4-Dichloroisocoumarin |
D7910 |
| Thrombin |
N-Acetyl-eglin C |
E7888 |
| Thrombin |
Gabexate mesylate |
G2417 |
| Thrombin |
[Lys47]-Hirudin (HV2), leech |
H0393 |
| Thrombin |
Hirudin from leeches |
H7016 |
| Thrombin |
Hirudin from leeches |
H7380 |
| Thrombin |
Hirudin from leeches |
H9022 |
| Thrombin |
Leupeptin trifluoroacetate salt |
L2023 |
| Thrombin |
Na-(2-Naphthalenesulfonylglycyl)-4-amidino-DL-phenylalaninepiperidide |
N8514 |
| Thrombin |
Phenylmethanesulfonyl fluoride |
P7626 |
| Thrombin |
Na-p-Tosyl-L-lysine chloromethyl ketone hydrochloride |
T7254 |
| Thrombin |
D-Val-Leu-Lys-chloromethyl ketone |
V3763 |
| Thromboxane A2 synthase |
Ozagrel |
O1385 |
| Thromboxane synthase |
Picotamide |
P8477 |
| Thromboxane synthase |
Furegrelate sodium salt |
F3764 |
| Thromboxane synthase |
Picotamide |
P8477 |
| Thromboxane synthase |
Furegrelate sodium salt |
F3764 |
| Tissue Plasminogen Activator |
a2-Antiplasmin from human plasma |
A0914 |
| Tissue Plasminogen Activator |
Aprotinin from bovine lung |
A1153 |
| Topoisomerase I |
Apigenin |
A3145 |
| Topoisomerase I |
Kaempferol |
K0133 |
| Topoisomerase I |
b-Lapachone |
L2037 |
| Topoisomerase I |
Rebeccamycin from Saccharothrix aerocolonigenes |
R4900 |
| Topoisomerase II |
(-)-Arctigenin |
A1854 |
| Topoisomerase II |
Aurintricarboxylic acid ammonium salt |
A0885 |
| Topoisomerase II |
Amsacrine hydrochloride |
A9809 |
| Topoisomerase II |
Chrysomycin A ³98% (HPLC) solid |
C6616 |
| Topoisomerase II |
Chrysomycin B ³98% (HPLC) solid |
C6491 |
| Topoisomerase II |
Ellipticine |
E3380 |
| Topoisomerase II |
Sobuzoxane |
S4692 |
| Topoisomerase II-B |
XK469 ³98 % (HPLC) solid |
X3628 |
| TrK A Kinase |
GW441756 hydrochoride powder, red ³97% (HPLC) |
G3420 |
| Trypsin |
a2-Antiplasmin from human plasma |
A0914 |
| Trypsin |
Aprotinin from bovine lung |
A1153 |
| Trypsin |
a1-Antitrypsin from human plasma |
A6150 |
| Trypsin |
Antipain hydrochloride |
A6191 |
| Trypsin |
4-Aminobenzamidine dihydrochloride |
A7148 |
| Trypsin |
Antithrombin III from human plasma |
A2221 |
| Trypsin |
4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride |
A8456 |
| Trypsin |
a1-Antitrypsin from human plasma |
A9024 |
| Trypsin |
Antithrombin III from bovine plasma |
A9141 |
| Trypsin |
Bdellin from leeches |
B3906 |
| Trypsin |
Benzamidine hydrochloride hydrate |
B6506 |
| Trypsin |
Diisopropyl fluorophosphate |
D0879 |
| Trypsin |
3,4-Dichloroisocoumarin |
D7910 |
| Trypsin |
Ecotin |
E1398 |
| Trypsin |
Gabexate mesylate |
G2417 |
| Trypsin |
Leupeptin trifluoroacetate salt |
L2023 |
| Trypsin |
a2-Macroglobulin from human plasma |
M6159 |
| Trypsin |
Phenylmethanesulfonyl fluoride |
P7626 |
| Trypsin |
Trypsin inhibitor from bovine pancreas |
T0256 |
| Trypsin |
Trypsin inhibitor from chicken egg white |
T2011 |
| Trypsin |
Trypsin inhibitor from turkey egg white |
T4385 |
| Trypsin |
Na-p-Tosyl-L-lysine chloromethyl ketone hydrochloride |
T7254 |
| Trypsin |
Trypsin inhibitor Glycine max (soybean) |
T9008 |
| Trypsin |
Trypsin inhibitor from Glycine max (soybean) |
T9128 |
| Trypsin |
Trypsin inhibitor from chicken egg white |
T9253 |
| Trypsin |
Trypsin inhibitor from Phaseolus limensis (lima bean) |
T9378 |
| Trypsin |
Trypsin-chymotrypsin inhibitor from Glycine max (soybean) |
T9777 |
| Tyramine oxidase |
p-Benzoquinone |
B1266 |
| Urokinase |
Glu-Gly-Arg-chloromethyl ketone |
G2917 |