Order Center
Biochemicals
Antibiotic Explorer
BioUltra
Biomaterials
Detection
Detergent Theory
Enzyme Explorer
BioUltra Proteins
Product Indexes
Cell Signaling Enzymes
Analytical Enzymes
Enzyme Reagents
Key Resources
Enzyme Literature
Assay Library
Carbohydrate Analysis
Cell Detachment
Collagenase Guide
Cytochrome C
Enzyme & Inhibitor Guide
Lysing Enzymes
Nucleases
Plasma and Blood Protein
Protease Finder
Proteolytic Enzymes
Protease Inhibitors
Protein Kinase Explorer
Lipids
Microparticles
Pharmacopoeia
PolyAmino Acids
Reference Chart
BioFiles
 Enzymes for Cell Detachment and Tissue Dissociation
Enzyme Explorer
Select the ideal enzyme for your cell detachment or tissue dissociation needs with these helpful links. Learn about the factors that influence your tissue digestion-dissociation and discover optimal conditions for working with these enzymes.
Collagenase
Because of its unique ability to hydrolyze native collagen, it is widely used to isolate cells from animal tissue.
Hyaluronidase
Catalyzes the random hydrolysis of
1,4-β-D-glycosidic linkages.
Papain
Relatively nonspecific sulfhydryl protease that can be used alone for dissociation.
Elastase
Useful for the dissociation of tissues containing higher amounts of elastin.
DNase
Reduces viscosity from DNA released from damaged cells during harvesting.
Protease Type XIV
Extremely nonspecific protease for dissociation of various tissues.
Trypsin
Serine protease commonly used for cell detachment and tissue dissociation.