|
Prealbumin, thyroxine-binding
Retinol-binding protein
Albumin
α-Globulins
β-Globulins
Apolipoproteins
γ-Mobility (non-immunoglobulin)
Complement Components
Coagulation Proteins
Cell-Related Plasma Proteins
Immunoglobulins (Antibodies)
Amyloid Proteins (Non-Immunoglobulin)
Misc. Trace Components
Plasma & Blood Protein Resource
| Prealbumin, thyroxine-binding |
| Protein | Species | Subunit Structure | Method | Chain Length | Protein MW | Amino Terminus | Carb. Content (%) |
| Prealbumin, thyroxine-binding | Human | Noncovalent tetramer of identical chains | AA | 127 | 54,980 | Gly | 0 |
| Retinol-binding protein | back to top |
| Protein | Species | Subunit Structure | Method | Chain Length | Protein MW | Amino Terminus | Carb. Content (%) |
| Retinol-binding protein | Human | Single chain | AA | 182 | 20,957 | Glu | 0 |
| Albumin | back to top |
| Protein | Species | Subunit Structure | Method | Chain Length | Protein MW | Amino Terminus | Carb. Content (%) |
| Albumin | Human | Single chain | AA | 585 | 66,458 | Asp | 0 |
| | | Nucleic acid precursor | NA | 609 | 69,365 | | 0 |
| | Bovine | Single chain | AA | 582 | 66,210 | Asp | 0 |
| | | Nucleic acid precursor | NA | 606 | | | |
| | Porcine | Single chain | AA | ~575 | ~66,300 | Asp | 0 |
| | Ovine | Single chain | AA | 575 | ~66,300 | Asp | 0 |
| | Chicken | Single chain | AA | 575 | ~66,300 | Asp | 0 |
| | Rat | Single chain | AA | 584 | 64,600 | Glu | 0 |
| | | Nucleic acid precursor | NA | 608 | | Glu | |
| α-Globulins |
back to top |
| Protein | Species | Subunit Structure | Method | Chain Length | Protein MW | Amino Terminus | Carb. Content (%) |
| α-Globulins | | | | | | | |
| α1-Microglobulin | Human | Single chain | AA | 167 | 26,000 | Gly | 20 |
| α1-Acid glycoprotein | Human | Single chain | AA | 181 | 40,000 | pGlu | 42 |
| | Rat | Nucleic acid precursor | NA | 187 | ~40,000 | Gln | + |
| α1-Antitrypsin | Human | Single chain | AA | 394 | 54,000 | Glu | 13 |
| | | Nucleic acid precursor | NA | | | | |
| | Baboon | Nucleic acid precursor | NA | 394 | ~54,000 | Glu | + |
| α1-Antichymortrypsin | Human | Single chain | AA | ~440 | 68,000 | Asn | 26 |
| | | | NA | 408 | | | |
| α2-Antiplasmin | Human | Single chain | AA | | 65,000 | Asn | 11 |
| α1-Fetoprotein | Human | Nucleic acid precursor | NA | 590 | 66,300 | Thr | 3 |
| | Rat | Nucleic acid precursor | NA | | | | |
| | Mouse | Nucleic acid precursor | NA | 586 | 72,000 | Ala | |
9.5 S α1-Glycoprotein (serum amyloid P component) | Human | Two pentagons (10 non-covalently bound identical subunits) | AA | (182)10 | ~250,000 | His | 13 |
| α1B-Glycoprotein | Human | Possibly a single chain | AA | ~480 | 68,000 | Ala | 13 |
| α1T-Glycoprotein | Human | Single chain | | ~776 | 85,000 | | 15 |
Vitamin D-binding protein (Gc globulin) | Human | Single chain | AA | ~450 | 52,000 | Leu | 4 |
| Thyroxine-binding protein | Human | Single chain | | | 54,000 | | 20 |
| Inter-α-trypsin inhibitor | Human | Single chain | AA | | ~160,000 | Lys | 8 |
| Ceruloplasmin | Human | Single chain (easily cleaved) | AA | 1,046 | 132,000 | Lys | 7 |
| 3.8 S Histidine-rich α2-glycoprotein | Human | Single chain | AA | ~440 | ~58,500 | Val | 11 |
| Galactoglycoprotein | Human | Single chain | AA | ~213 | 81,000 | Blocked | 76 |
| 3.1 S Leucine-rich α2-glycoprotein | Human | Single chaine | AA | 312 | 50,000 | Val | 23 |
| α2µ-Globulin | Rat (male) | Nucleic acid precursor | NA | 162 | 18,709 | Glu | + |
| Zn-α2-Glycoprotein | Human | Single chain | | ~258 | 41,000 | | 18 |
| α2HS-Glycoprotein | Human | Two chains (A & B) | AA | ~359 | 49,000 | Ala | 14 |
| | | B chain | AA | 27 | 3,386 | Thr | + |
| α2-macroglobulin | Human | 4 identical subunits | AA | 1,450 | 725,000 | Ser | 9.4 |
| 4S-α2,β1-Glycoprotein | Human | Single chain | | ~465 | 60,000 | Arg | 28 |
| Haptoglobin | Human | 2 pairs of nonidentical chains (αβ)2 | AA | 656 | 86,018 | Val/Ile | 16.4 |
| | | Single-chain precursor | NA | 245 | | | |
| | | α1 chain | AA | 83 | 9,189 | Val | 0 |
| | | α2 chain | AA | 142 | 15,939 | Val | 0 |
| | | β chain | AA | 245 | 33,820 | Ile | 19.4 |
| | | β chain | NA | | | | |
| | Canine | One pair of chains (αβ) β chain | AA | | | | |
| Corticosteroid-binding globulin | Human | Single chain | | | 52,000 | | 30 |
| 8 S α3-Glycoprotein | Human | Tetramer | | | 220,000 | | 31 |
| β-Globulins |
back to top |
| Protein |
Species |
Subunit Structure |
Method |
Chain Length |
Protein MW |
Amino Terminus |
Carb. Content (%) |
| Pregnancy-specific β1-glycoprotein Human | A trimeric aggregate | | | (42,300)n | | 32.6 |
| Transferrin | Human | Single chain | AA | 678 | 79,550 | Val | 5.9 |
| | | | NA | 679 | | Val | |
| Ovotransferrin | Chicken | Single chain | AA | 686 | 77,770 | Ala | + |
| | | Nucleic acid precursor | NA | 686 | | | |
| Hemopexin | Human | Single chain | AA | 439 | 60,000 | Thr | 22 |
| | Rabbit | Single chain | AA | ~440 | 57,000 | Val | 20 |
| β2-Microglobulin | Human | Single chain | AA | 99 | 11,731 | Ile | 0 |
| | Mouse | Single chain | AA | 99 | | Ile | 0 |
| | Rabbit | Single chain | AA | 99 | | Val | 0 |
| | Guinea pig | Single chain | AA | 99 | | Val | 0 |
| C-Reactive protein | Human | Subunits aggregate as cyclic pentamers | AA | 187 | (20,946)5 | pGlu | 0 |
| | Rabbit | Pentagonal structure | AA | 186 | | Ala | 0 |
| β2-Glycoprotein I | Human | Single chain | AA | 326 | ~50,000 | Gly | 19 |
| β2-Glycoprotein III | Human | Single chain, forms complex | AA | | 35,000 | | 10 |
| Apolipoproteins |
back to top |
| Protein |
Species |
Subunit Structure |
Method |
Chain Length |
Protein MW |
Amino Terminus |
Carb. Content (%) |
| Apolipoprotein A-I (HDL) | Human | Single chain; no disulfides | AA | 243 | 28,076 | Asp | 0 |
| Apolipoprotein A-II (HDL) | Human | Dimer of 2 identical chains, joined by disulfide | AA | 77 | 17,414 | pGlu | 0 |
| Apolipoprotein C-I (VLDL) | Human | Single chain; no disulfides | AA | 57 | 6,631 | Thr | 0 |
| Apolipoprotein C-II | Human | Single chain; no disulfides | AA | 78 | 8,829 | Thr | 0 |
| Apolipoprotein C-III (VLDL) | Human | Single chain glycopeptide | AA | 79 | 8,764 | Ser | 3 |
| Apolipoprotein E | Human | Single chain | AA | 299 | 34,183 | Lys | + |
| γ-Mobility (non-immunoglobulin) |
back to top |
| Protein |
Species |
Subunit Structure |
Method |
Chain Length |
Protein MW |
Amino Terminus |
Carb. Content (%) |
| 0.6 S γ2-Globulin | Human | Single chain | AA | 47 | 5,100 | Phe | 0 |
| 2 S γ2-Globulin | Human | Single chain | AA | 132 | 14,000 | Tyr | 0 |
| Basic Protein B2 | Human | Single chain | AA | ~85 | 9,000 | | 0 |
| Post-γ-Globulin (γ-trace) | Human | Single chain | AA | 120 | 13,248 | Ser | 0 |
| Complement Components |
back to top |
| Protein |
Species |
Subunit Structure |
Method |
Chain Length |
Protein MW |
Amino Terminus |
Carb. Content (%) |
| C1q | Human | Six identical nocovalently-linked subunits, each with three similar chains (A,B&C) Total of 18 chains | AA | | 410,000 | Glu/Asn | 9 |
| | | B chain | AA | 226 | | Glu | + |
| | | C chain | AA | ~200 | | Asn | + |
| C1r | Human | Noncovalent dimer | | | (83,000)2 | | 9.4 |
| C1r | Human | B chain | AA | 242 | 27,096 | Ile | 12.4 |
| C1s | Human | Single chain | AA | | 83,000 | Glu | 7.1 |
| C1s | Human | Two chains (a and b) | AA | | 83,000 | Glu/Ile | 7.1 |
| C2 | Human | Single chain | AA | | 102,000 | Ala | + |
| C3 | Human | Two disulfide-bonded chains | AA | ~1,630 | ~185,000 | | 2 |
| | | α chain | AA | ~960 | 115,000 | Ser | |
| | | β chain | AA | ~670 | 75,000 | Ser | |
| | Mouse | ProcC3 is a single-chain precursor | NA | | ~190,000 | Ile | |
| C3a | Human | Fragment of C3 a chain | AA | 77 | 9,093 | Ser | 0 |
| | Porcine | Fragment of C3 α chain | AA | 77 | ~9,000 | Ser | |
| | Rat | Fragment of C3 α chain | AA | 78 | ~9,000 | Ser | |
| | Mouse | Fragment of C3 α chain | NA | 78 | ~9,800 | Ser | |
| C4 | Human | Three disulfide-bonded chains | AA | | | 7 | |
| | | α chain | AA | ~785 | 93,000 | | |
| | | β chain | AA | ~660 | 75,000 | | |
| | | γ chain | AA | ~250 | 33,000 | | |
| C4a | Human | Fragment of a chain | AA | 77 | 8,763 | Asn | 0 |
| | Bovine | Fragment of a chain | AA | 77 | | | |
| C5 | Human | Two disulfide-bonded chains | | | 185,000 | | 1.6 |
| | | α chain | AA | ~960 | 115,000 | Thr | |
| | | β chain | | ~670 | 75,000 | | |
| C5a | Human | Fragment of α chain | AA | 74 | 8,274 | Thr | + |
| C6 | Human | Single chain | AA | ~900 | 1,048,000 | Ala | 3.8 |
| C7 | Human | Single chain | AA | ~800 | 92,400 | Ser | 6.4 |
| C8 | Human | Two noncovalently-linked subunits | | | 163,000 | | |
| C4-Binding Protein | Human | Disulfide-bonded subunits (Mr 75,000) | | | 540,000 | Asn | |
| Factor D (C3 Proactivator Convertase) | Human | Single chain | AA | | 24,000 | Ile | + |
| Factor B (C3 Proactivator) | Human | Single chain | AA | | 90,000 | Thr | 6 |
| | | Bb Fragment | AA | 505 | 60,000 | Lys | + |
| Properdin | Human | Four non-covalent identical chains | AA | ~480 | 220,000 | Asp | 10 |
| Factor H (β1H) | Human | Single chain | AA | ~1,275 | 155,000 | Glu | 9.3 |
| C3b-Inactivator | Human | Two disulfide-bonded chains | | | 90,000 | Ile/Lys | 11 |
| | | Light chain | AA | | 40,000 | Ile/Lys | + |
| | | Heavy chain | AA | | 50,000 | Lys | + |
| Coagulation Proteins |
back to top |
| Protein |
Species |
Subunit Structure |
Method |
Chain Length |
Protein MW |
Amino Terminus |
Carb. Content (%) |
| Prothrombin | Human | Single chain | AA | 581 | 72,000 | Ala | 12 |
| | Bovine | Single chain | AA | 582 | 72,000 | Ala | 12 |
| Thrombin | Bovine | Two disulfide-linked chains | AA | ~340 | 37,000 | Thre/ILE | |
| | | A chain | AA | 49 | 5,728 | Thre/ILE | 0 |
| | | B chain | AA | 259 | 31,000 | Ile | |
| FactorVII | Human | Single chain (two chains when activated) | AA | | 48,000 | Ala | 16 |
| Factor VIII | Human | Noncovalent complex | | | (100,000)n | | 6 |
| Factor IX | Human | Single chain Precursor | NA | 462 | 51,801 | | + |
| | Bovine | Single chain | AA | 427 | ~55,000 | Tyr | 26 |
| Factor IXa (activated) | Human | Heavy and light chains | NA | 381 | | | |
| | | Light chain | NA | 145 | | Tyr | |
| | | Heavy chain | AA | 236 | | Val | |
| | Bovine | Heavy and light chains | AA | 390 | ~46,500 | Tyr/Val | 11 |
| | | Light chain | AA | 146 | 16,600 | Tyr | 7.4 |
| | | Heavy chain | AA | 232 | 27,300 | Val | 13 |
| Factor X | Human | Heavy and light chains | AA | | 59,000 | Ala/Ser | 15 |
| | | Light chain | AA | 139 | 16,211 | Ala/Ser | 0 |
| | Bovine | Heavy and light chains | AA | 447 | 55,000 | Trp/Ala | 10 |
| | | Light chain | AA | 140 | 16,143 | Ala/Ser | 0 |
| | | Heavy chain | AA | 307 | 38,000 | Trp/Ala | 11.5 |
| Factor Xa Activated | Bovine | Heavy and light chains | AA | 396 | 44,000 | Ile/Ala | 3 |
| | | Light chain | AA | 140 | 16,000 | Ala | 0 |
| | | Heavy chain | AA | 256 | 28,000 | Ile | 0 |
| Factor XI | Human | Two idenical disulfide-linked chains | AA | | 130,000 | Gly | 5 |
| Protein C | Bovine | Two disulfide-linked chains | AA | 415 | 55,000 | Ala/Asp | 14 |
| | | Light chain | AA | 155 | 21,000 | Ala | + |
| | | Heavy chain | AA | | |