Order Center
Biochemicals
Antibiotic Explorer
BioUltra
Biomaterials
Detection
Detergent Theory
Enzyme Explorer
BioUltra Proteins
Product Indexes
Cell Signaling Enzymes
Analytical Enzymes
Enzyme Reagents
Key Resources
Enzyme Literature
Assay Library
Carbohydrate Analysis
Cell Detachment
Collagenase Guide
Cytochrome C
Enzyme & Inhibitor Guide
Lysing Enzymes
Nucleases
Plasma and Blood Protein
Protease Finder
Proteolytic Enzymes
Protease Inhibitors
Protein Kinase Explorer
Lipids
Microparticles
Pharmacopoeia
PolyAmino Acids
Reference Chart
BioFiles
 Protease Inhibitor Specificity Index I
Enzyme Explorer
Aminopeptidase
Angiotensin Converting Enzyme
APP-Secretase
b-Secretase
Bromelain
Calpains
Carboxypeptidases Caspases
Cathepsin
Chymotrypsins
Clostripain
Collagenase
Complement C1, C1r & C1s
Complement Factors B & D
Dipeptidyl peptidase IV
Dispase
Elastases
Endoproteinase Arg-C, Glu-C, Lys-C
Factor VIIIa, Xa, XIa, XIIa
Ficin
gSecretase
Granzyme B
HIV Protease
Kallikreins
Leucine Aminopeptidases
Matrix metalloprotease
Metalloprotease inhibitors
Papain
Pepsin
Plasmin
Pro-caspase 3
Pronase E
Proteases
Proteasome inhibitor
Proteinase K
Renins
Thermolysin
Thrombin
Trypsin
Urokinase
Aminopeptidase
Product Code Inhibitor
B8385 Bestatin
M6500 2-Mercaptoethylamine HCl
Aminopeptidase N
Product Code Inhibitor
P7240 Phebestin
Aminopeptidase P
Product Code Inhibitor
A1395 Apstatin
Angiotensin Converting Enzyme
Product Code Inhibitor
A0773 Angiotensin-converting enzyme inhibitor
B0507 Bradykinin potentiator B
B0632 Bradykinin potentiator C
C4042 Captopril
C8856 Captopril USP
F5017 Foroxymithine
F6800 Fenoldopam
S6633 N-Succinyl-L-proline
T7025 N-epsilon-Trifluoroacetyl-Lys-Pro
APP-Secretase
Product Code Inhibitor
A1847 [Asn670,Sta671,Val672]-Amyloid beta/A4 protein precursor 770 fragment (662-675)
back to top
b-Secretase
Product Code Inhibitor
G8291 Glu-Val-Asn-[(2R,4S,5S)-5-Amino-4-Hydroxy-2,7-Dimethylocanoyl]-Ala-Glu-Phe
S4562 b-Secretase Inhibitor
Bromelain
Product Code Inhibitor
E3132 E-64
E3876 N-Ethylmaleimide
I2512 Iodoacetic Acid
T7254 N-a-p-Tosyl-L-lysine chloromethyl ketone Hydrochloride
T4376 N-a-p-Tosyl-L-phenylalanine chloromethyl ketone Hydrochloride
Calpain
Product Code Inhibitor
A6191 Antipain
C4285 Acetyl-Calpastatin (184-210) Human
C5913 p-Chloromercurbenzoic acid Free Acid
C8984 Z-Leu-Leu-Leu-Fluoromethyl ketone
C9234 Z-Leu-Leu-Tyr-Fluoromethyl ketone
C9859 Z-Leu-Leu-Tyr-CHN2
E0514 (2S,3S)-trans-Epoxysuccinyl-/L-leucylamido-3-methylbutane
E3876 N-Ethylmaleimide
E8640 (2S,3S)-trans-Epoxysuccinyl-L-leucylamido-3-methylbutane ethyl ester
L2023 Leupeptin
M5439 MeOSuc-Val-homo-Phe-fluoromethyl ketone
M6159 a2-Macroglobulin
Calpain I
Product Code Inhibitor
A6060 N-Acetyl-Leu-Leu-Met-al
A6185 N-Acetyl-Leu-Leu-Norleu-al
C9181 Calpain Inhibitor Peptide
Calpain II
Product Code Inhibitor
A6060 N-Acetyl-Leu-Leu-Met-al
A6185 N-Acetyl-Leu-Leu-Norleu-al
A6191 Antipain
C4285 Acetyl-Calpastatin (184-210) Human
C5913 p-Chloromercuribenzoic acid Free Acid
C9181 Calpain Inhibitor Peptide
E0514 (2S,3S)-trans-Epoxysuccinyl-/L-leucylamido-3-methylbutane
E3876 N-Ethylmaleimide
L2023 Leupeptin
M6159 a2-Macroglobulin
back to top
Carboxypeptidase A
Product Code Inhibitor
C0279 Carboxypeptidase inhibitor
D6518 Diethylenetriaminepentaacetic acid Free Acid
ED2SS EDTA
E4378 EGTA
P9375 1,10-Phenanthroline Monohydrate
Carboxypeptidase B
Product Code Inhibitor
C0279 Carboxypeptidase inhibitor
D6518 Diethylenetriaminepentaacetic acid Free Acid
ED2SS EDTA
E4378 EGTA
P9375 1,10-Phenanthroline Monohydrate
Carboxypeptidase P
Product Code Inhibitor
C5913 p-Chloromercuribenzoic acid Free Acid
D5758 Deithyl pyrocarbonate
D0879 Diisopropyl fluorophosphate
I2512 Iodoacetic Acid
Carboxypeptidase Y
Product Code Inhibitor
A1153 Aprotinin
D0879 Diisopropyl fluorophosphate
P7626 Phenylmethylsulfonyl fluoride
Caspase
Product Code Inhibitor
B2057 Biotin-Val-Ala-Asp(OMe)-Fluoromethyl ketone
B2682 N-T-Boc-Asp(OMe)-fluoromethyl ketone
C8484 Z-Ala-Glu(OMe)-Val-Asp(OMe)-Fluoromethyl ketone
C8609 Z-Val-Lys(Biotin)-Asp(OMe)-Fluoromethyl ketone
Caspase 1
Product Code Inhibitor
A1466 N-Acetyl-Trp-Glu-His-Asp-al
A4211 N-Acetyl-Tyr-Val-Ala-Asp chloromethyl ketone
A4339 N-Acetyl-Tyr-Val-Lys(biotinyl)-Asp 2,6-dimethylbenzoyloxymethyl ketone
C9484 Z-Leu-Glu-(OMe)-Val-Asp(OMe)-Fluoromethyl ketone
Caspase 2
Product Code Inhibitor
A2222 N-Acetyl-Val-Asp-Val-Ala-Asp-CHO
C1605 N-Cbz-Val-Asp(OMe)-Val-Ala-Asp(OMe)-FMK
Caspase 3
Product Code Inhibitor
A0835 N-AcetylL-Asp-Glu-Val-Asp-al
A1216 N-Acetyl-Ile-Glu-Thr-Asp-al
A1341 N-Acetyl-Glu-Ser-Met-Asp-al
B1807 Biotin-Asp-(OMe)-Glu-(OMe)Val(OMe)-Asp(OMe)-Fluoromethyl ketone
C0480 N-Cbz-Asp(OMe)-Gln-Met-Asp(OMe)-FMK
C0859 Caspase-4 Inhibitor (aldehyde)
V116 Z-VAD-FMK
Caspase 5
Product Code Inhibitor
A1466 N-Acetyl-Trp-Glu-His-Asp-al
Caspase 6
Product Code Inhibitor
A6339 N-Acetyl-Val-Glu-Ile-Asp-al
B1807 Biotin-Asp-(OMe)-Glu-(OMe)Val(OMe)-Asp(OMe)-Fluoromethyl ketone
C0480 N-Cbz-Asp(OMe)-Gln-Met-Asp(OMe)-FMK
C1730 N-CBZ-Val-Glu-(OMe)-Ile-Asp(OMe)-FMK
Caspase 7
Product Code Inhibitor
B1807 Biotin-Asp-(OMe)-Glu-(OMe)Val(OMe)-Asp(OMe)-Fluoromethyl ketone
C0605 N-Cbz-Asp(OMe)-GLu(OMe)-Val-Asp(OMe)-FMK
Product Code Inhibitor
A1216 N-Acetyl-Ile-Glu-Thr-Asp-al
B1807 Biotin-Asp-(OMe)-Glu-(OMe)Val(OMe)-Asp(OMe)-Fluoromethyl ketone
C1230 N-Cbz-Ile-Glu(OMe)-Thr-Asp(OMe)
C8734 Z-Leu-Glu-(OMe)-Thr-Asp-(OMe)-Fluoromethyl ketone
Caspase 9
Product Code Inhibitor
C1355 N-Cbz-Leu-Glu(OMe)-His-Asp(OMe)-FMK
Caspase 10
Product Code Inhibitor
B1807 Biotin-Asp-(OMe)-Glu-(OMe)Val(OMe)-Asp(OMe)-Fluoromethyl ketone
Caspase 13
Product Code Inhibitor
C8859 Z-Leu-Glu(OMe)-Glu(OMe)-Asp(OMe)-Fluoromethyl ketone
back to top
Cathepsin B
Product Code Inhibitor
A6060 N-Acetyl-Leu-Leu-Met-al
A6185 N-Acetyl-Leu-Leu-Norleu-al
A6191 Antipain
B1932 Biotin-Phe-Ala-Fluoromethyl ketone
C0408 Cystatin
C5732 CA-074
C5857 CA-074 Methyl ester
C7268 Chymostatin
C8917 Cystatin
C9109 Z-Phe-Phe-Fluoromethyl ketone
E3132 E-64
L2023 Leupeptin
M5314 MeOSuc-Phe-homo-Phe fluoromethyl ketone
M6159 a2-Macroglobulin
P0350 Procathepsin B, rat, fragment 26-50
P0475 Procathepsin B, rat, fragment 36-50
Cathepsin C
Product Code Inhibitor
I2512 Iodoacetic Acid
Cathepsin D
Product Code Inhibitor
A6191 Antipain
C7268 Chymostatin
P5318 Pepstatin A
Cathepsin G
Product Code Inhibitor
A9285 a1-Antchymotrypsin
C7268 Chymostatin
C9984 Z-Gly-Leu-Phe-Chloromethyl ketone
D0879 Diisopropyl fluorophosphate
D7910 3,4-Dichloroisocoumarin
E7888 N-Acetyl-eglin C
Cathepsin L
Product Code Inhibitor
A6185 N-Acetyl-Leu-Leu-Norleu-al
C9109 Z-Phe-Phe-Fluoromethyl ketone
C9234 Z-Leu-Leu-Tyr-Fluoromethyl ketone
C9859 Z-Leu-Leu-Tyr-CHN3
M5314 MeOSuc-Phe-homo-Phe fluoromethyl ketone
back to top
back to Protease Inhibitors