|
Selecting an appropriate protein kinase inhibitor can pose a challenge due to the complexity of the protein kinase family. Factors such as inhibitor specificity and cell permeability should be considered. The table below is based on currently available data. For specific information on specific inhibitors, consult the citations. Specific kinetic data for inhibitors marked with an "X" was not available. References may be viewed by clicking in the reference column or scrolling down past the product table.
| Prod No. |
Ref No. |
Product Description |
PKA |
PKC |
PKG |
CAM Kinase |
MLCK |
Casein Kinase |
PTK |
Notes & MAPK, CDK, Other Kinases |
|
A165 | 1-5 | Adenosine 3',5'-cyclic-Monophosphothioate, RP-Isomer |
11µM | | | |
| | | |
| A7410 |
6 | Amiloride HCl | | |
| | | |
EGFR | |
|
A3145 |
7-9 | Apigenin | | 10µM | |
| | | FGFR(20µM) | |
|
A8186
| 10 |
Arg-Lys-Arg-Ala-Arg-Lys-GLu | 350-460µM |
| 85-100µM | | |
| | |
|
A4308 |
11 | Autocamtide 2-Related Inhibitory Peptide | | | | CAMKII(40nM) | | | | |
|
B6292
| 12 | Bisindolylmaleimide I Hydrochloride (GF 109203X) | 2.0µM | 14nM | | | | | | |
|
B178 |
13-15 | Butein | >500µM | >500µM | | | | | See Notes | EGFR(16µM); p60c-src(65µM) |
|
P1431 |
18-19 | Calmodulin | | | | |
| | | GRK 2&3(2µM) GRK5(40nM) & GRK6 |
|
C6303 | 20-21 | Calphostin C | >50µM | 50nM | | | | | >50µM |
|
|
C2932
| 23 | Chelerythrine chloride | | 0.66µM |
| | | | | |
|
C240 |
24 | Rp-8-[(4Chlorophenyl)thio]-cGMPS Triethylamine | | | 0.5µM | |
| | | |
| D7802
| 25 | Daidzein | | |
| |
| CKII | | Does not inhibit MAP2Kinase |
|
D3768
| 27 | Dequalinium Chloride | | 8-15µM |
| | | | | β1 (8-15µM), I,II,III (11µM) |
|
D1916 | 29 | 5,6-Dichlorobenzimidazole riboside | | | |
| | CKII(4-10µM) | | |
|
D4681 |
30-32 | L-threo-Dihydrosphingosine | | X |
| | | | | |
|
D2667 | 33,34 |
2,5-Dihydroxycinnamic Acid Methyl Ester | | |
| | | | EGFR |
|
|
E2250 |
36 | Ellagic Acid | 2µM | 8µM | | |
| | | |
|
E7881 | 37-39
| Emodin | >200µM | >200µM |
| | | CKII(2µM) |
| |
|
E3645 |
40 | rac-2-Ethoxy-3-Hexadecanamido-1-Propyl Phosphocholine | |
X | | | | | | ID50=2.32µM for Leukemic HL60 cells |
|
E3770
| 40 | rac-2-Ethoxy-3-Octadecanamido-1-Propyl Phosphocholine | |
X | | | | |
| ID50=1.64µM for Leukemic HL60 cells |
|
G3381
| 41-43 | Geldanamycin | |
| | |
| | erbB, EGFR, v-src | |
G6649 G6776 | 44-50 | Genistein | >100µM | 15µM
| | | | | See Notes | EGFR(2.6µM)insulin(41µM)V-src(26µM) V-abl(39µM) |
|
G0897 |
51 | Genistin | |
| | |
| | Neg. Control | |
|
G2911
| 12 | GF 109203X (Bisindolylmaleimide I Hydrochloride) | 2.0µM | 10nM |
| | | | | |
|
I7016
| 52,53 | H-7 Dihydrochloride
(1-(5-Isoquinolinesulfonyl)-2-methylpiperazine Dihydrochloride) | 3.0µM | 6.0µM | 5.8µM | | 97µM | CKI(100µM) KII(780µM) | | |
|
I6891 |
52,53 | H-7 Free Base (1-(5-Isoquinolinesulfonyl)-2-methylpiperazine)
| 3.0µM | 6.0µM | 5.8µM | | 97µM |
CKI(100µM) CKII(780µM) | | |
|
M9656
| 52 | H-8, N-(2-[Methylamino]ethyl)-5-isoquinolinesulfonamide Hydrochloride | 1.2µM |
14.4µM | 0.48µM | | 68µM |
CKI(133µM) CKII(950µM) | | |
|
B1427 |
54-57 | H-89 |
48nM | 14µM | 340nM | CAMKII(11µM) | 39µM | CKI(34µM), CKII(124µM) |
| |
|
I1392
| 59 | HA-100 |
8µM | 12µM | 4µM | |
240µM | | | |
|
G1274
| 59-65 | HA-1004 | 2.3µM | 40µM | 1.3µM |
CAMKII(13µM) | 150µM | | |
|
|
H139 |
66 | HA-1077 2HCL | 1.6µM |
| 1.6µM | | 36µM | | | |
|
H6649 | 67-69 | Herbimycin A
| | | | | | |
p210bcr-abl | |
|
H5257
| 71 | Hispidin |
| X | | | | | | |
|
H7904 |
72 | Hydroxytamoxifen | | 88µM | | | | | | |
|
H9252 | 73,74
| Hypericin | >50µM | 3.3µM |
| | | | | |
|
I0404 | 75
| Indirubin 3'-monoxime | | |
| | | | | Glycogen synthase Kinase
(5-50nM)&
CDK(5-100nM) |
|
I2142 | 76-82 | KN-62 |
>100µM | >100µM |
| CAMKII(0.3-1.8µM) | >100µM |
| | |
| K112 |
81-86 | KN-92 |
| | | Neg.Control |
| | | |
|
K1385 | 83 | KN-93 |
| | | | | |
| |
|
K3761 | 87 | KT5720 |
60nM | | 2.4nM | | |
| | |
|
L2400 | 88-91 | Lavendustin A |
| | | |
| | EGFR(4.4ng/ml) | |
|
L5025 | 92
| Leflunomide | | | | |
| | BTK | |
|
M2272 | 93,94 | Melittin | |
1-4µM | |
| 0.08-1.4µM | | |
|
| M7795
| 40 | rac-2-Methoxy-3-Hexadecanamido-1-Propyl
phosphocholine | |
X | | | | | | |
|
M8045 | 40 | rac-2-Methoxy-3-Octadecanamido-1-Propyl
phosphocholine | |
X | | | | |
| |
|
I2764 | 95-97 |
ML-7 |
21µM | 42µM | | |
300nM | | | |
|
C1172 | 97-103 |
ML-9 | | | | X | X |
| | |
|
M6760 | 104-105
| Myricetin | 27.5µM | 12.1µM |
| | 1.7µM | CKI(9µM), CKII(0.6µM) |
pp130fps(1.8µM), insulin receptor(2.6) | |
|
N161 |
106 | NPC-15437 DiHCl |
| X | | | |
| | |
|
O0886 | 107 | Olomoucine |
| | | |
| | | CDK Inhibitor |
|
P4509 | 108-109 |
Palmitoyl-DL-Carnitine Chloride | | 34µM | | | |
| | |
|
P215 |
110-112 | PD 098,059 |
| | | | | |
| prevents the activation of MAPKK1 by Raf or MEK kinase |
|
P7912 | 113 |
Phloretin | | ~5µM | |
| | | | |
|
P0453 | 114-118 | Piceatannol
| 3µM | 8µM | |
| 12µM | |
See notes | Tyrosine kinases Syk, p40(15µM), p56lck(66µM) and p72. CDK(19µM) |
|
P1004 | 119 | Polymyxin B Sulfate
| | ~10µM | |
| | | | |
|
P6062 | 120 |
Protein kinase A inhibitor fragment 6-22 amide | 1.6nM | |
| | | | | |
| P8462 |
121 | Protein Kinase C Fragment 19-36 | 423µM | 147nM | | | 24µM | | |
|
|
P0393 | 122-125 | Protein Kinase Inhibitor
from porcine heart | X | | |
| | | | |
|
P0300 | 123-125 | Protein Kinase Inhibitor
Rabbit Sequence | 2.3nM | | |
| | | | |
|
P5015 | 123-125 | Protein Kinase Inhibitor from rabbit
muscle | X | | |
| | | | |
|
Q0125 | 105,126, 127 | Quercetin | | 20-40µM | | | | CKG(220nM), CKI(45µM), CKII(100nM) | See Notes | EGF Receptor(410nM), I, IIA, IIB(10µM), III(10µM), Mg++ Dep., P60v-src(50µM) |
|
R2146 | 128-133 |
Radicicol | | | | |
| | src(0.1µg/ml) | |
|
R0395 | 134-135 | Rapamycin | | | | |
| | p70s6k, p33cdk2 and p34cdc2. |
mTOR |
| R136 |
136-139 | Ro 31-8220 (Bisindolylmaleimide IX) | 1.5µM
| 7.6nM | |
CAMKII(17µM) | | | | GRK5, GSK3
|
|
R137 |
136,140, 141 | Ro 32-0432 |
| | | |
| | | 10-fold selectivity for PKCα; 4-fold selectivity for PKCβI over PKCε. GRK5 inhibitor; less potent inhibitor of GRK2 or GRK3 |
| R7772 | 142,143 | Roscovitine |
| | | |
| | | p34cdc2(0.45µM), CDK2 |
|
R5648 | 144
| Rottlerin | | See notes | |
CAMKIII(5.3µM) | | | | PKCδ 3-6 µM; PKCα, PKCβ, PKCγ 30-42 µM |
|
S8307 | 145 | SB 203580 | | | | | |
| | p38 MAPK (0.5µM) |
| S7049
| 146-148 | D-Sphingosine |
| 53µM | |
CAMKIII(20µM) | | | | Also Inhibits MAPK |
| S4400 |
73,105, 149-155 | Staurosporine | 6nM |
2.7-5.5nM | 8.5nM | CAMKII(20nM) |
21nM | | C-src(10nM), EGFR(600nM), Insulin(60nM)V-Src(6nM)V-abl(80nM) |
Inhibits CDK 1,2&5 |
| T5648 |
157,158 | Tamoxifen Free Base | | 5-15µM | | | | | | |
| T9262 | 157,158 | Tamoxifen Citrate | | 5µM | |
| | | | |
|
T3126 | 160 | (+)-a-Tocopherol Acid Succinate | | X |
| | | | | |
|
T7254 |
105 | N a-p-Tosyl-L-lysine
chloromethyl ketone Hydrochloride | |
1mM | | | | | | |
|
T7040
| 161 | Tyrphostin 1 |
| | | | | | EGFR >1250µM | Negative Control |
|
T7165 |
161 | Tyrphostin 23 | | | |
| | | EGFR 35µM | |
| T7665 |
161 | Tyrphostin 51 | |
| | | |
| EGFR 0.8µM | |
| T7790
| 161 | Tyrphostin 63 | |
| | | |
| EGFR 6500µM | Negative Control |
| T182 |
162,163 | Tyrphostin A9 |
| | | | | |
PDGFR(0.5µM), EGFR(460µM) | |
| T9177
| 164 | Tyrphostin AG 126 | | | | | | | X | Blocks TNFα and NO production in macrophages |
|
T3434 |
51,165,
166 | Tyrphostin AG 490 | |
| | | | | Jak-2 | |
| T2067
| 167 | Tyrphostin AG 879 | | | | | | | pp140c-trk(10µM) | |
| T4057 |
168 | Tyrphostin AG 1296 | |
| | | |
| PDGFR | |
|
T5317 | 161 | Tyrphostin AG 1433 |
| | | | |
| PDGR & VEGFR | PDGFβ receptor(5 µM); VEGFreceptor(9.3µM) |
| T4182
| 169 | Tyrphostin AG 1478 | |
| | | |
| EGFR | |
|
T4192 | 161 | Tyrphostin SU 1498 |
| | | | |
| VEGFR(700nM) | |
| U120 |
170-172 | U0126 |
| | | | | |
| MEK1(72nM), MEK2(58nM) |
|
W1628 | 173,174
| Wortmannin | | | | |
200nM | | | PI3Kinase(3nM) |
| No. |
References |
| 1 | Rothermel, J.D., et al., A mechanistic and kinetic analysis of the interactions of the diastereoisomers of adenosine 3',5'-(cyclic)phosphorothioate with purified cyclic AMP-dependent protein kinase, Biochem. J., 251, 757 (1988). |
| 2 | Parker Botelho, L.H., et al., cAMP analog antagonists of cAMP action, Methods Enzymol., 159, 159-172 (1988). |
| 3 | Richter-Landsberg, C., Jastorff, B., In vitro phosphorylation of microtubule-associated protein 2: Differential effects of cyclic AMP analogues, J. Neurochem., 45, 1218-1222 (1985). |
| 4 | Dostmann, W.R.G., et al., Probing the cyclic nucleotide binding sites of cAMP-dependent protein kinases I and II with analogs of adenosine
3',5'-cyclic phosphorothioates, J. Biol. Chem., 265, 10484 (1990). |
| 5 | Yokozaki, H., et al., Unhydrolyzable analogues of adenosine 3':5'-monophosphate demonstrating growth inhibition and differentiation in human cancer cells, Cancer Res., 52, 2504 (1992). |
| 6 | Davis, R.J., Czech, M.P., Amiloride directly inhibits growth factor receptor tyrosine kinase activity, J. Biol. Chem., 260, 2543-51 (1985) |
| 7 | Huang, Y.T., et al., Inhibitions of protein kinase C and proto-oncogene expressions in NIH 3T3 cells by apigenin, Eur. J. Cancer, 32, 146-151 1996). |
| 8 | Kuo ML, Yang NC, Reversion of v-H-ras-transformed NIH 3T3 cells by apigenin through inhibiting mitogen activated protein kinase and
its downstream oncogenes.Biochem. Biophys. Res. Commun., 212, 767-75 (1995) |
| 9 | Yin F.M, Signal pathways involved in apigenin inhibition of growth and induction of apoptosis of human anaplastic thyroid cancer cells. Anticancer Res., 19, 4297-303 (1999) |
| 10 | Glass, D., et al., Differential and common recognition of the catalytic sites of the cGMP-dependent and cAMP-dependent protein kinases by
inhibitory peptides derived from the heat-stable inhibitor protein, Biochem. J., 213, 159 (1983) |
| 11 | Ishida, A., et al., A novel highly specific and potent inhibitor of calmodulin-dependent protein kinase II., Biochem. Biophys. Res. Commun., 212, 806-12 (1995) |
| 12 | Toullec, D., et al., The isindolylmaleimide GF 109203X is a potent and selective inhibitor of protein kinase C., J. Biol. Chem., 266, 15771 (1991). |
| 13 | Yang, E., et al., Butein, a specific protein tyrosine kinase inhibitor, Biochem. Biophys. Res. Commun., 245, 435-438 (1998). |
| 14 | Yu, S.M., et al., Endothelium-
dependent relaxation of rat aorta by butein, a novel cyclic AMP-specific phosphodiesterase inhibitor,
Eur. J. Pharmacol., 280, 69-77 (1995). |
| 15 | Aizu, E., et al., Inhibition of 12-O-tetradecanoylphorbol-13-acetate-mediated epidermal ornithine decarboxylase induction and skin tumor promotion by new lipoxygenase inhibitors lacking protein kinase C inhibitory effects, Carcinogenesis, 7, 1809-1812 (1986). |
| 16 | Kitagawa, M., et al., Butyrolactone I, a selective inhibitor of cdk2 and cdc2 kinase., Oncogene, 8, 2425-32 (1993) |
| 17 | Okuyama, A., et al., [CDK and MMP inhibitors]., Gan To Kagaku Ryoho, 24, 1547-62 (1997) |
| 18 | Chuang, T.T., et al., Inhibition of G protein-coupled receptor kinase subtypes by Ca2+/calmodulin., J. Biol. Chem., 271, 28691-6 (1996) |
| 19 | Pitcher, J. A., Freedman, N. J. and Lefkowitz, R. J., G protein-coupled receptor kinases., Ann. Rev. Biochem,. 67, 653-92 (1998). |
| 20 | Tamaoki, T. and Nakano, H., Potent and specific inhibitors of protein kinase C of microbial origin, Biotechnology (NY), 8, 732-735 (1990). |
| 21 | Kobayashi, E., et al., Calphostin C (UCN-1028C), a novel microbial compound, is a highly potent and specific inhibitor of protein kinase
C. Biochem. Biophys. Res. Commun., 159, 548-553 (1989). |
| 22 | Smith, et al., Functional determinants in the autoinhibitory domain of calcium/calmodulin-dependent protein kinase II. J. Biol. Chem., 267, 1761-1768 (1992) |
| 23 | Herbert, J.M., et al., Chelerythrine is a potent and specific inhibitor of protein kinase C. Biochem. Biophys. Res. Commun., 172, 993-999 (1990). |
| 24 | Butt, E., (Rp)-8-pCPT-cGMPS, a novel cGMP-dependent protein kinase inhibitor., Eur J Pharmacol., 269, 265-8 (1994) |
| 25 | Higashi, K. and Ogawara, H., Daidzein inhibits insulin- or insulin-like growth factor-1-mediated signaling in cell cycle progression of Swiss
3T3 cells., Biochim. Biophys. Acta., 1221, 29-35 (1994) |
| 26 | Faltyrek, C.R., et al., Damnacanthal is a highly potent, selective inhibitor of p56lck tyrosine kinase activity. Biochemistry, 34, 12404
(1995). |
| 27 | Rotenberg, S.A., Inhibition of rodent protein kinase C by the anticarcinoma agent dequalinium., Cancer Res., 50, 677-85 (1990) |
| 28 | Yen, A., Scoong, S., Retinoic acid-induced RB (retinoblastoma) hypophosphorylation enhanced by CGP 52411 (4,5-dianilinophthalimide), an EGF family tyrosine kinase receptor inhibitor. Eur. J. Cell Biol., 69, 327-334 (1996). |
| 29 | Zandomeni, R., et al., Casein kinase type II is involved in the inhibition by 5,6-dichloro-1-beta-D-ribofuranosylbenzimidazole of specific RNA polymerase II transcription, J. Biol. Chem., 261, 3414-9 (1986) |
| 30 | Schwartz, G.K., et al., A pilot clinical/pharmacological study of the protein kinase C-specific inhibitor safingol alone and in combination
with doxorubicin., Clin. Cancer Res., 3, 537-43 (1997). |
| 31 | Schwartz, G.K., et al., Potentiation of apoptosis by treatment with the protein kinase C-specific inhibitor safingol in mitomycin C-treated
gastric cancer cells., J. Natl. Cancer Inst., 87, 1394-9 (1995) |
| 32 | Sachs, C.W., et al., Partial inhibition of multidrug resistance by safingol is independent of modulation of P-glycoprotein substrate
activities and correlated with inhibition of protein kinase C., J. Biol. Chem., 270, 26639-48 (1995) |
| 33 | Geltz, et al., The p85 and p110 subunits of phosphatidylinositol 3-kinase-alpha are substrates, in vitro, for a constitutively associated
protein tyrosine kinase in platelets. Blood, 91, 930-939 (1998). |
| 34 | Haier, et al., Influence of phosphotyrosine kinase inhibitors on adhesive properties of highly and poorly metastatic HT-29 colon carcinoma cells to collagen. Int. J. Colorectal. Dis., 14, 119-127 (1999). |
| 35 | Basson, et al., Effects of modulation of tyrosine phosphorylation on brush border enzyme activity in human Caco-2 intestinal epithelial
cells. Cell. Tissue Res., 292, 553-562 (1998). |
| 36 | Wang, B.H., et al., Inhibition of eukaryote serine/threonine-specific protein kinases by piceatannol., Planta Med., 64, 195-9 (1998) |
| 37 | Yim, et al., Emodin, an anthraquinone derivative isolated from the rhizomes of Rheum palmatum, selectively inhibits the activity of casein
kinase II as a competitive inhibitor. Planta Med., 65, 9-13 (1999). |
| 38 | Kumar, et al., Emodin (3-methyl-1,6,8-trihydroxyanthraquinone) inhibits TNF-induced NF-kappaB activation, IkappaB degradation, and expression of cell surface adhesion proteins in human vascular endothelial cells. Oncogene, 17, 913-918 (1998). |
| 39 | Zhang, et al., Tyrosine kinase inhibitor emodin suppresses growth of HER-2/neu-overexpressing breast cancer cells in athymic mice and
sensitizes these cells to the inhibitory effect of paclitaxel. Clin. Cancer Res., 5, 343-353 (1999). |
| 40 | Marx, M.H., et al., Synthesis and evaluation of neoplastic cell growth inhibition of 1-N-alkylamide analogues of glycero-3-phosphocholine.,
J. Med. Chem., 31, 858 (1988) |
| 41 | Whitesell, L., et al., Stable and specific binding of heat shock protein 90 by geldanamycin disrupts glucocorticoid receptor function in intact cells. Mol. Endocrinol., 10, 705-712 (1996). |
| 42 | Supko, J.G., et al., Preclinical pharmacologic evaluation of geldanamycin as an antitumor agent. Cancer Chemother. Pharmacol., 36, 305-315 (1995). |
| 43 | Chavany, C., et al., p185erbB2 binds to GRP94 in vivo. Dissociation of the p185erbB2/GRP94 heterocomplex by benzoquinone ansamycins precedes depletion of p185erbB2. J. Biol. Chem., 271, 4974-4947 (1996). |
| 44 | Geissler, J.F., Thiazolidine-diones. Biochemical and biological activity of a novel class of tyrosine protein kinase inhibitors., J. Biol. Chem., 265, 22255-61 (1990) |
| 45 | Akiyama, T., et al., Genistein, a specific inhibitor of tyrosine-specific protein kinases. J. Biol. Chem., 262, 5592-5595 (1987). |
| 46 | Koroma, B.M. and de Juan, E., Inhibition of protein tyrosine phosphorylation in endothelial cells: relationship to antiproliferative action of genistein., Biochem. Soc. Trans., 25, 35-40 (1997) |
| 47 | O'Dell, T.J., et al., Long-term potentiation in the hippocampus is blocked by tyrosine kinase inhibitors. Nature, 353, 558-560 (1991). |
| 48 | Huang J., et al., Genistein inhibits protein Histidine kinase. J. Biol. Chem., 267, 15511-15515 (1992). |
| 49 | Hakansson, G., and Allen, J.F., Histidine and tyrosine phosphorylation in pea mitochondria: evidence for protein phosphorylation in respiratory
redox signalling. FEBS Lett., 372, 238-242 (1995). |
| 50 | Igarashi, M., and Komiya, Y., Tyrosine phosphorylation and immunodetection of vinculin in growth cone particle (GCP) fraction and in CP-cytoskeletal Subfractions.J. Neurosci. Res., 30, 266-274 (1991). |
| 51 | Potier, et al., Protein tyrosine kinase inhibitors reduce high-activating calcium currents in CA1 pyramidal neurones from rat hippocampal slices.
Brain Res., 816, 587-597 (1999). |
| 52 | Bradshaw, D., et al, Therapeutic potential of protein kinase C inhibitors., Agents Actions 1993 Jan;38(1-2):135-47 |
| 53 | Hidaka, H., et al., Isoquinolinesulfonamides, novel and potent inhibitors of cyclic nucleotide dependent protein kinase and protein kinase C. Biochemistry, 23, 5036-5041 (1984). |
| 54 | Chijiwa, T., et al., J. Biol. Chem., 265, 5267 (1990) |
| 55 | Hidaka, et al., The Biology and Medicine of Signal Transduction, pp. 485-90, Nishizuka, Y., et al., eds, Raven Press NY (1990) |
| 56 | Murol, M. and Suzuki, T., Cell Signal., 5, 289 (1993) |
| 57 | Fujihara, M., et al., J. Biol. Chem., 268, 14898 (1993) |
| 58 | Inagaki, et al., N-(2-Aminoethyl)-5-isoquinolinesulfonamide, a newly synthesized protein kinase inhibitor, functions as a ligand in affinity chromatography. J. Biol. Chem., 260, 2922(1985). |
| 59 | Hagiwara, M., et al., Selective modulation of calcium-dependent myosin phosphorylation by novel protein kinase inhibitors, isoquinolinesulfonamide derivatives., Mol Pharmacol., 32, 7-12 (1987) |
| 60 | Xiao, G.H., et al., cAMP-dependent negative regulation of rat aldehyde dehydrogenase class 3 gene expression. J. Biol. Chem., 272, 3238-3245
(1997). |
| 61 | Luo, S.F., et al., Effect of forskolin on bradykinin-induced calcium mobilization in cultured canine tracheal smooth muscle cells. Cell
Signal., 9, 159-167 (1997). |
| 62 | Svenningsen, A.F. and Kanje, M., Regulation of Schwann cell proliferation in cultured segments of the adult rat sciatic nerve. J. Neurosci.
Res., 52, 530-537 (1998). |
| 63 | Crowley, M.R., et al., HA1004, an intracellular calcium antagonist, selectively attenuates pulmonary hypertension in newborn lambs. J.
Cardiovasc. Pharmacol., 23, 806-813 (1994). |
| 64 | Leahy, J.C. and Vallano, M.L., Differential effects of isoquinolinesulfonamide protein kinase inhibitors on CA1 responses in hippocampal slices. Neuroscience, 44, 361-370 (1991). |
| 65 | Tsushima, H., et al., Human erythropoietin receptor increases GATA-2 and Bcl-xL by a protein kinase C-dependent pathway in human erythropoietin-dependent cell line AS-E2. Cell Growth Differ., 8, 1317-1328 (1997). |
| 66 | Asano, T., et al., Vasodilator actions of HA1077 in vitro and in vivo putatively mediated by the inhibition of protein kinase., Br J Pharmacol., 98, 1091-100 (1989) |
| 67 | Fukazawa, H., et al., Specific inhibition of cytoplasmic protein tyrosine kinases by herbimycin A in vitro., Biochem. Pharmacol., 42, 1661-71(1991) |
| 68 | Zakar, et al., Tyrosine kinase inhibitors block the glucocorticoid stimulation of prostaglandin endoperoxide H synthase expression in
amnion cells. Can. J. Physiol. Pharmacol., 77, 138-142 (1999). |
| 69 | Davis, et al., Herbimycin A and geldanamycin inhibit okadaic acid-induced apoptosis and p38 activation in NRK-52E renal epithelial cells. Toxicol. Appl. Pharmacol., 16, 59-74 (1999). |
| 70 | Uberall, F., et al., Hexadecylphosphocholine inhibits inositol phosphate formation and protein kinase C activity., Cancer Res., 51, 807-12 (1991) |
| 71 | Gonindard, C., et al., Synthetic hispidin, a PKC inhibitor, is more cytotoxic toward cancer cells than normal cells in vitro., Cell Biol.
Toxicol., 13, 141-53 (1997) |
| 72 | Eyster, K.M., Nonsteroidal antiestrogen inhibition of protein kinase C in human corpus luteum and placenta., Biochem. Pharmacol., 38, 3497-503
(1989) |
| 73 | Tamaoki, T., et al., The Biology and Medicine of Signal Transduction, pp. 497-501, Nishizuka, Y., et al., eds, Raven Press NY (1990) |
| 74 | Takahashi, I., et al., Hypericin and pseudohypericin specifically inhibit protein kinase C: possible relation to their antiretroviral activity. Biochem. Biophys. Res. Commun., 165, 1207-1212 (1989). |
| 75 | Leclerc, S., Indirubins Inhibit Glycogen Synthase Kinase-3beta and CDK5/P25, Two Protein Kinases Involved in Abnormal Tau Phosphorylation
in Alzheimer's Disease., J. Biol. Chem., 276, 251-260 (2001) |
| 76 |
Minami, H., The effect of KN-62, Ca2+/calmodulin dependent protein kinase II inhibitor on cell cycle., Biochem. Biophys. Res. Commun., 199, 241-8 (1994) |
| 77 | MacLeod, et al., Ca(2+)/Calmodulin kinase II and decreases in intracellular pH are required to activate K(+) channels after substantial swelling in villus epithelial cells. J. Membr. Biol., 172, 59-66 (1999). |
| 78 | Fan, et al., Inhibition of calcium/calmodulin-dependent protein kinase II in rat hippocampus attenuates morphine tolerance and dependence. Mol. Pharmacol., 56, 39-45 (1999). |
| 79 | Communi, et al., Calcium-calmodulin-dependent protein kinase II and protein kinase C-mediated phosphorylation and activation of D-myo-inositol 1,4, 5-trisphosphate 3-kinase B in astrocytes. J. Biol. Chem., 274, 14734-14742 (1999). |
| 80 | Fan, et al., Inhibition of calcium/calmodulin-dependent protein kinase II in rat hippocampus attenuates morphine tolerance and dependence. Mol. Pharmacol., 56, 39-45 (1999). |
| 81 | Sumi, M., et al., The newly synthesized selective Ca2+/calmodulin dependent protein kinase II inhibitor KN-93 reduces dopamine contents
in PC12h cells., Biochem. Biophys. Res. Commun., 181, 968-75 (1991) |
| 82 | Tokumitsu, H., et al., KN-62, 1-[N,O-bis(5-isoquinolinesulfonyl)-N-methyl-L-tyrosyl]-4-phenylpiperazine, a specific inhibitor of 2+/calmodulin-dependent protein kinase II. J. Biol. Chem., 265, 4315-20 (1990) |
| 83 | Tessier, et al., Regulation of the transient outward K(+) current by Ca(2+)/Calmodulin-dependent protein kinases II in human atrial
myocytes. Circ. Res., 85, 810-819 (1999). |
| 84 | Tombes, R.M., et al., G1 cell cycle arrest and apoptosis are induced in NIH 3T3 cells by KN-93, an inhibitor of CaMK-II (the multifunctional
Ca2+/CaM kinase)., Cell Growth Differ., 6, 1063-70 (1995) |
| 85 | Smith, et al., Increased calcium/calmodulin protein kinase activity in astrocytes chronically exposed to ethanol: influences on glutamate transport.Neurosci. Lett., 269, 145-148 (1999). |
| 86 | Wasmeier, et al., Secretagogue-dependent phosphorylation of phogrin, an insulin granule membrane protein tyrosine phosphatase homologue. Biochem. J., 341, 563-569 (1999). |
| 87 | Kase, H., K-252 compounds, novel and potent inhibitors of protein kinase C and cyclic nucleotide-dependent protein kinases., Biochem
Biophys. Res. Commun., 142, 436-40 (1987) |
| 88 | Hsu, C.Y., et al, Kinetic analysis of the inhibition of the epidermal growth factor receptor tyrosine kinase by Lavendustin-A and its analogue.
J. Biol. Chem., 266, 21105-12 (1991) |
| 89 | Onoda, T., Isolation of a novel tyrosine kinase inhibitor, lavendustin A, from Streptomyces griseolavendus., J. Nat. Prod., 52, 1252-7 (1989) |
| 90 | Huang, et al., Protein tyrosine kinase is required for the induction of long-term potentiation in the rat hippocampus. J. Physiol. (Lond),
520, 783-796 (1999). |
| 91 | Taskinen, et al., Inhibition of atrial wall stretch-induced cardiac hormone secretion by lavendustin A, a potent tyrosine kinase inhibitor.
Endocrinology, 140, 4198-4207 (1999). |
| 92 | Mahajan, S., et al., Rational design and synthesis of a novel anti-leukemic agent targeting Bruton's tyrosine kinase (BTK), LFM-A13 [alpha-cyano-beta-hydroxy-beta-methyl-N-(2, 5-dibromophenyl)propenamide]., J. Biol. Chem., 274, 9587-99 (1999) |
| 93 | Katoh, N., et al., Inhibition by melittin of phospholipid-sensitive and calmodulin-sensitive Ca2+-dependent protein kinases. Biochem.
J., 202, 217-24 (1982) |
| 94 | Raynor, R.L., et al., Membrane interactions of amphiphilic polypeptides mastoparan, melittin, polymyxin B, and cardiotoxin. Differential inhibition of protein kinase C, Ca2+/calmodulin-dependent protein kinase II and
synaptosomal membrane Na,K-ATPase, and Na+ pump and differentiation of HL60 cells., J. Biol. Chem., 266, 2753 (1991) |
| 95 | Krarup. T., et al., Na+-K+-2Cl- cotransport in Ehrlich cells: regulation by protein phosphatases and kinases. Am. J. Physiol., 275, 239-50
(1998) |
| 96 | Makishima, M., et al., Induction of differentiation of human leukemia cells by inhibitors of myosin light chain kinase., FEBS Lett., 287,
175 (1991). |
| 97 | Saitoh, M., et al., Selective inhibition of catalytic activity of smooth muscle myosin light chain kinase. J. Biol. Chem., 262, 7796-7801 (1987) |
| 98 | Saitoh, M., et al., The modulatory role of myosin light chain phosphorylation in human platelet activation., Biochem. Biophys. Res. Commun., 140,
280-7 (1986) |
| 99 | Hidaka, H., Properties and use of H-series compounds as protein kinase inhibitors., Methods Enzymo., 201, 328-39 (1991) |
| 100 | Ito, M., et al., Inhibition of natural killer cell-mediated cytotoxicity by ML-9, a selective inhibitor of myosin light chain kinase. Int.
J. Immunopharmacol., 11, 185-190 (1989). |
| 101 | Inoue, G., et al., Effects of ML-9 on insulin stimulation of glucose transport in 3T3-L1adipocytes. J. Biol. Chem., 268, 5272-5278 (1993). |
| 102 | Nagatsu, T., Effects of myosin light-chain kinase inhibitor on catecholamine secretion from rat pheochromocytoma PC12h cells. Biochem. Biophys. Res. Commun., 143, 1045-8 (1987) |
| 103 | Reig, J.A., et al, Naphthalenesulfonamide derivatives ML9 and W7 inhibit catecholamine secretion in intact and permeabilized chromaffin cells. Neurochem. Res., 18, 317-323 (1993). |
| 104 | Hagiwara, M., et al., Differential effects of flavonoids as inhibitors of tyrosine protein kinases and serine/threonine protein kinases.
Biochem Pharmacol., 37, 2987-92 (1988) |
| 105 |
Handbook of Enzyme Inhibitors, H. Zollner (1993) |
| 106 | Sullivan, et al., 2,6-Diamino-N-([1-(1-oxotridecyl)-2-piperidinyl]-methyl)hexanamide (NPC 15437): A selective inhibitor of protein kinase C. Agents Actions, 34, 138-141 (1991). |
| 107 | Vesely, J., et al., Inhibition of cyclin-dependent kinases by purine analogues. Eur. J. Biochem., 224, 771-786 (1994). |
| 108 | Nakadate, T., and Blumberg, P.M., Modulation by palmitoylcarnitine of protein kinase C activation. Cancer Res., 47, 6537-6542 (1987) |
| 109 | Robinson, P.J., et al., MDL 27,032 [4-propyl-5-(4-pyridinyl)-2(3H)-oxazolone], an active site-directed inhibitor of protein kinase C and cyclic AMP-dependent protein kinase that relaxes vascular smooth muscle., J. Pharmacol. Exp. Ther., 255, 1392-8 (1990) |
| 110 | Alessi, et al., PD 098059 is a specific inhibitor of the activation of mitogen-activated protein kinase kinase in vitro and in vivo. J. Biol. Chem., 270, 27489-27494 (1995). |
| 111 | Dudley, et al., A synthetic inhibitor of the mitogen-activated protein kinase cascade. Proc. Natl. Acad. Sci. USA, 95, 7686-7689 (1995) |
| 112 | Pang, et al., Inhibition of MAP kinase blocks the differentiation of PC-12 cells induced by nerve growth factor. J. Biol. Chem., 270,
13585-13588 (1995). |
| 113 | Grosman, N., Inhibitory effect of phloretin on histamine release from isolated rat mast cells., Agents Actions, 25, 284-90 (1988) |
| 114 | Wang, B.H., Panta Med., 64, 195 (1998) |
| 115 | Cambien, et al., Src-regulated extracellular signal-related kinase and syk-regulated c-Jun N-terminal kinase pathways act in conjunction
to induce IL-1 synthesis in response to microtubule disruption in HL60 cells. J. Immunol., 163, 5079-85 (1999). |
| 116 | Geahlen, R.L., Piceatannol (3,4,3',5'-tetrahydroxy-trans-stilbene) is a naturally occurring protein-tyrosine kinase inhibitor.Biochem.
Biophys. Res. Commun., 165, 241-5 (1989) |
| 117 | Tkaczyk, et al., Fc epsilon RI-mediated antigen endocytosis turns interferon-gamma-treated mouse mast cells from inefficient into potent
antigen-presenting cells. Immunology, 97, 333-340 (1999). |
| 118 | Zheng, et al., Piceatannol, a stilbene phytochemical, inhibits mitochondrial F0F1-ATPase activity by targeting the F1 complex. Biochem. Biophys.
Res. Commun., 261, 499-503 (1999). |
| 119 | Nel, A.E., Polymyxin B causes coordinate inhibition of phorbol ester-induced C-kinase activity and proliferation of B lymphocytes., Biochem. Biophys. Res. Commun., 128, 1364-72 (1985) |
| 120 | Glass, D.B., et al., Protein kinase inhibitor-(6-22)-amide peptide analogs with standard and nonstandard amino acid substitutions for
phenylalanine 10. Inhibition of cAMP-dependent protein kinase., J. Biol. Chem., 264, 14579 (1989). |
| 121 | House, C. and Kemp, B.E., Protein Kinase C contains a pseudosubstrate protope in its regulatory domain., Science, 238, 1726 (1987) |
| 122 | Brostrom, C.O. and Kon, C., An improved protein binding assay for cyclic AMP. Anal. Biochem., 58, 459 [1974]. |
| 123 | |