Sigma-Aldrich
Sigma - Life Science Logo
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Enzyme Explorer
Tyrosine Kinases
Receptor Linked
Non-Receptor Linked
Overview
Protein Kinase Tables Home
Tyrosine Kinases — Receptor Linked
Abbreviations
Currently
Accepted
Name
EGFR
(E 3641 (human),
E 2645 (human))
HER-2/Neu/cerbB2
PDGFR
IGF1-R
FGFR
(F 2897)
Ligands
EGF
(E 9644 (human),
E 4127 (m))
TGFalpha (T 7924 (human),
T 9533 (r))
Amphiregulin
(A 7080 (human))
Unknown
PDGF
(P 3201 (BB) (human),
P 3076 (AA) (human))
IGF-1
(I 3769 (human))
FGF
(F 3133 (bovine),
F 5542 (a) (human),
F 0291 (b) (human))
Structural
Information
1210 aa (human)
1255 aa (human)
1098alpha (human)
1106beta (human)
1367 aa (human)
822 aa (mouse)
Mechanism
of Activation
Ligand-induced dimerization
followed by
trans-auto-
phosphorylation
Probably by hetero-
dimerization
Dimerization of
the receptor
followed by
trans-auto-
phosphorylation
Trans-auto-
phosphorylation
within the dimer
Dimerization of
the receptor
followed by
trans-auto-
phosphorylation
Selective
Inhibitors
AG 1478
(T 4182)
PD 153,035
AG 825
AG 879
(T 2067)
AG 1295
(T 3932)
AG 1296
(T 4057)
AGL 2043
AG 1024
AG 538
SU 5402
Non-Selective
Inhibitors
DAPH 1
(D-210)
Lavendustin A
(L 2400)
Genistein
(G 6649,
G 6776)
Herbimycin A
(H 6649)
DAPH 1
(D-210)
Lavendustin A
(L 2400)
Genistein
(G 6649,
G 6776)
Herbimycin A
(H 6649)
Lavendustin A
(L 2400)
Genistein
(G 6649,
G 6776)
Herbimycin A
(H 6649)
Not tested
SU 4984
Diseases
Implicated
Gliomas, squamous cell carcinoma, papilloma, psoriasis
Breast and ovarian carcinomas, non-small cell lung cancer
Glioblastomas, atherosclerosis, restenosis
Prostate cancer, breast cancer
Radioligands
of Choice
125I]-EGF
[125I]-hPDGF
[125I]-IGF-1
[125I]-FGF
Abbreviations
AG 538: 3,4-Dihydroxy[ (3',4'-dihydroxy)benzoyl]-cis-cinnamonitrile
AG 825: 4-Hydroxy-3-methoxy-5-(benzothiazoylthiomethyl)benzylidenecyanoacetamide
AG 879: alpha-Cyano-(3,5-di-t-butyl-4-hydroxy)thiocinnamide
AG 1024: 3-Bromo-5-t-butyl-4-hydroxybenzylidenemalonitrile
AG 1295: 6,7-Dimethyl-2-phenylquinoxaline
AG 1296: 6,7-Dimethoxy-2-phenylquinoxaline
AG 1478: N-(3-Chlorophenyl)-6,7-dimethoxy-4-quinazolinamine
AGL 2043: 1,2-Dimethyl-6-(2-thiophene)imidazolo[5,4-g]-quinoxaline
DAPH 1: 4,5-Dianilinophthalimide
EGF: Epidermal growth factor
FGF: Fibroblast growth factor
IGF-1: Insulin-like growth factor-1
PD 153,035: 4-(3-Bromoanilino)-6,7-dimethoxyquinazoline
PDGF: Platelet-derived growth factor
SU 5402: 3-[3-(2-Carboxyethyl)-4-methylpyrrol-2-methylidenyl]-2-indolinone
SU 4984: 3-[4-(1-Formylpiperazin-4-yl)benzylidenyl]-2-indolinone
TGF: Transforming growth factor
a: acidic
b: basic
m: mouse
r: rat
back to top
Tyrosine Kinases — Non-Receptor Linked
Abbreviations
Currently
Accepted
Name
pp60 c-Src and src family kinases
Bcr-Abl
Jak-2 and Jak family kinases
Ligands
None
None
None
Structural
Information
542 aa (mouse)
a
1133 aa (human)
Mechanism
of Activation
Dephosphorylation of a C-terminal phosphotyrosine
Dimerization
Recruitment followed by conformational change
Selective
Inhibitors
PP1
PP2
7,8-DHIC
AG 1112
AG 957
Non-Selective
Inhibitors
Herbimycin A (H 6649)
AG 18
AG 34
AG 82
Herbimycin A (H 6649)
Herbimycin A (H 6649)
Diseases
Implicated
In its viral form (v-Src) cancer in chicken; activated in certain human cancers (c-Src)
Bcr-Abl causes chronic myeloid leukemia a
Pre-B recurrent acute lymphoblastic leukemia
Abbreviations
AG 18: alpha-Cyano-(3,4-dihydroxy) cinnamonitrile
AG 34: 3,4-Dihydroxy-5-methoxybenzylidine malononitrile
AG 82: alpha -Cyano-(3,4,5-trihydroxy)cinnamonitrile
AG 957: 4-Amino-N-(2,5-dihydroxybenzyl)methylbenzoate
AG 1112: 3-Indolidene malononitrile [-(3-amino-4-cyano-5-yl)]pyrazole
7,8-DHIC: 7,8-Dehydroxy-isoquinoline-3-carboxamide
PP1: 4-Amino-5-(4-methylphenyl)-7-(t-butyl)pyrazolo[3,4-d]pyrimidine
PP2: 4-Amino-5-(4-chlorophenyl)-7-(t-butyl)pyrazolo[3,4-d]pyrimidine
Footnotes
a Bcr-Abl is a chimeric protein consisting of the amino terminal 927 amino acids or the amino terminal 426 amino acids of Bcr fused to the second exon of the tyrosine kinase Abl.
back to top