Order Center
Biochemicals
Antibiotic Explorer
BioUltra
Biological Detergents
Detection
Enzyme Explorer
BioUltra Proteins
Product Indexes
Cell Signaling Enzymes
Analytical Enzymes
Enzyme Reagents
Key Resources
Enzyme Literature
Assay Library
Carbohydrate Analysis
Cell Detachment
Collagenase Guide
Cytochrome C
Enzyme & Inhibitor Guide
Lysing Enzymes
Nucleases
Plasma and Blood Protein
Protease Finder
Proteolytic Enzymes
Protease Inhibitors
Protein Kinase Explorer
Lipids
Microparticles
Peptide Explorer
Pharmacopoeia
PolyAmino Acids
Achromopeptidase, E.C. 3.4.21.50
Product Name
Product # 
Achromopeptidase from Achromobacter lyticus
A3547
Achromopeptidase from Achromobacter lyticus, Crude powder, 300-600 units/mg solid
A7550
Achromopeptidase from Achromobacter lyticus, Partially purified powder, 20,000-40,000 units/mg solid
A3422
Specificity:
Preferential cleavage: Lys-Xaa, including -Lys-Pro-
back to top
back to Proteolytic Enzymes Guide
Aminopeptidase, E.C. 3.4.11.10
Product Name
Product # 
Aminopeptidase from Aeromonas proteolytica
A8200
Inhibitors:
2-Mercaptoethylamine HCl, M6500
back to top
back to Proteolytic Enzymes Guide
Ancrod, E.C. 3.4.21.74
Product Name
Product # 
Ancrod from Agkistrodon rhodostoma (Malayan pit viper) venom
A5042
back to top
back to Proteolytic Enzymes Guide
Angiotensin Converting Enzyme, E.C.3.4.15.1
Product Name
Product # 
Angiotensin Converting Enzyme from porcine kidney, Lyophilized powder, minimum 10 units/mg protein (Bradford)
A2580
Angiotensin Converting Enzyme from rabbit lung, Lyophilized powder, 2-6 units/mg protein (modified Warburg-Christian)
A6778
Specificity:
Conversion of angiotensin I to angiotensin II
Synthethic Substrates:
N-[3-(2-Furyl)acryloyl]-Phe-Gly-Gly, F7131
N-Hippuryl-His-Leu tetrahydrate, H1635
Hippuryl-Gly-Gly, H1763
Hippuryl-His-Leu acetate salt, H4884
Inhibitors:
Angiotensin-converting enzyme inhibitor, A0773
Bradykinin potentiator B, B0507
Bradykinin potentiator C, B0632
Captopril, C4042
Captopril USP, C8856
Foroxymithine, F5017
Fenoldopam, F6800
N-Succinyl-L-proline, S6633
N-e-Trifluoroacetyl-Lys-Pro, T7025
back to top
back to Proteolytic Enzymes Guide
Bromelain, E.C. 3.4.22.32
Product Name
Product # 
Bromelain pineapple stem, Lyophilized powder 5-10 units/mg protein B5144
Bromelain pineapple stem, 3-7 units/mg protein B4882
Specificity:
Broad specificity for cleavage of proteins, but strong preference for Z-Arg-Arg NHMec in peptides
Synthethic Substrates:
N-a-CBZ-L-Lysine-p-Nitrophenyl ester, C3637
Inhibitors:
E-64, E3132
N-Ethylmaleimide, E3876
Iodoacetic Acid, I2512
N-a-p-Tosyl-L-lysine chloromethyl ketone Hydrochloride, T7254
N-a-p-Tosyl-L-phenylalanine chloromethyl ketone Hydrochloride, T4376
back to top
back to Proteolytic Enzymes Guide
Calpain
Inhibitors:
Antipain, A6191
Acetyl-Calpastatin (184-210) Human, C4285
p-Chloromercurbenzoic acid Free Acid, C5913
Z-Leu-Leu-Leu-Fluoromethyl ketone, C8984
Z-Leu-Leu-Tyr-Fluoromethyl ketone, C9234
Z-Leu-Leu-Tyr-CHN2, C9859
(2S,3S)-trans-Epoxysuccinyl-/L-leucylamido-3-methylbutane, E0514
N-Ethylmaleimide, E3876
(2S,3S)-trans-Epoxysuccinyl-L-leucylamido-3-methylbutane ethyl ester, E8640
Leupeptin , L2023
MeOSuc-Val-homo-Phe-fluoromethyl ketone, M5439
a2-Macroglobulin, M6159
back to top
back to Proteolytic Enzymes Guide
Calpain I, E.C. 3.4.22.52
Product Name
Product # 
Calpain 1 Human C6108
Specificity:
Broad endopeptidase specificity
Synthethic Substrates:
N-Succinyl-Leu-Tyr 7-amido-4-methylcoumarin, S1153
Inhibitors:
N-Acetyl-Leu-Leu-Met-al, A6060
N-Acetyl-Leu-Leu-Norleu-al, A6185
Calpain Inhibitor Peptide, C9181
back to top
back to Proteolytic Enzymes Guide
Calpain II, E.C. 3.4.22.32
Product Name
Product # 
Calpain II from rabbit skeletal muscle P4533
Specificity:
Preferential cleavage where P1', P2' and P3' are hydrophobic residues
Synthethic Substrates:
N-Succinyl-Leu-Tyr 7-amido-4-methylcoumarin, S1153
N-Succinyl-Leu-Leu-Val-Tyr-7-Amido-4-Methylcoumarin, S6510
Inhibitors:
N-Acetyl-Leu-Leu-Met-al, A6060
Acetyl-Calpastatin (184-210) Human, C4285
p-Chloromercuribenzoic acid Free Acid, C5913
Calpain Inhibitor Peptide, C9181
(2S,3S)-trans-Epoxysuccinyl-/L-leucylamido-3-methylbutane, E0514
N-Ethylmaleimide, E3876
Leupeptin, L2023
a2-Macroglobulin, M6159
back to top
back to Proteolytic Enzymes Guide
Carboxypeptidase A, E.C. 3.4.22.17
Product Name
Product # 
Carboxypeptidase A-Agarose
C1261
Carboxypeptidase A bovine pancreas, Type I Aqueous suspension approx. 50 units/mg protein
C0261
Carboxypeptidase A, PMSF treated bovine pancreas
C9268
Carboxypeptidase A bovine pancreas, Aqueous suspension approx. 50 units/mg protein
C0386
Specificity:
Release of a C-terminal amino acid, but little or no action with -Asp, -Glu, -Arg, -Lys or -Pro
Synthethic Substrates:
Hippuryl-L-phenylalanine, H6875
Inhibitors:
Carboxypeptidase inhibitor, C0279
Diethylenetriaminepentaacetic acid Free Acid, D6518
EDTA, ED2SS
EGTA, E4378
1,10-Phenanthroline Monohydrate, P9375
back to top
back to Proteolytic Enzymes Guide
Carboxypeptidase B, E.C. 3.4.17.2
Product Name
Product # 
Carboxypeptidase B from human pancreas C5233
Specificity:
Preferential release of a C-terminal lysine or arginine amino acid
Synthethic Substrates:
Hippuryl-L-Lys, H6750
Hippuryl-L-Arg, H2508
Inhibitors:
Carboxypeptidase inhibitor, C0279
Diethylenetriaminepentaacetic acid Free Acid, D6518
EDTA, ED2SS
EGTA, E4378
1,10-Phenanthroline Monohydrate, P9375
back to top
back to Proteolytic Enzymes Guide
Carboxypeptidase G, E.C. 3.4.19.9
Product Name
Product # 
Carboxypeptidase G from Pseudomonas sp. C9658
Specificity:
Hydrolysis of a gamma-glutamyl bond
Synthethic Substrates:
(+)-Amethopterin, A6770
back to top
back to Proteolytic Enzymes Guide
Carboxypeptidase P, E.C. 3.4.17.16
Product Name
Product # 
Carboxypeptidase P from Penicillium janthinellum C5396
Specificity:
Release of a C-terminal residue other than proline, by preferential cleavage of a prolyl bond
Synthethic Substrates:
N-CBZ-Glu-Tyr, C0257
Inhibitors:
Diisopropyl fluorophosphate, D0879
Iodoacetic Acid, I2512
back to top
back to Proteolytic Enzymes Guide
Carboxypeptidase W, E.C. 3.4.16.1
Product Name
Product # 
Carboxypeptidase W from wheat C6527
Synthethic Substrates:
N-CBZ-Phe-L-Ala, C1634
back to top
back to Proteolytic Enzymes Guide
Carboxypeptidase Y, E.C. 3.4.16.5
Product Name
Product # 
Carboxypeptidase Y from Baker's yeast (S. cerevisiae), Lyophilized powder minimum 50 units/mg protein C3888
Carboxypeptidase Y from Baker's yeast (S. cerevisiae), Lyophilized powder C4046
Specificity:
Release of a C-terminal amino acid with broad specificity
Synthethic Substrates:
N-CBZ-Phe-L-Ala, C1634
Inhibitors:
Aprotinin, A1153
Diisopropyl fluorophosphate, D0879
Phenylmethylsulfonyl fluoride, P7626
back to top
back to Proteolytic Enzymes Guide