Order Center
Biochemicals
Antibiotic Explorer
BioUltra
Biological Detergents
Detection
Enzyme Explorer
BioUltra Proteins
Product Indexes
Cell Signaling Enzymes
Analytical Enzymes
Enzyme Reagents
Key Resources
Enzyme Literature
Assay Library
Carbohydrate Analysis
Cell Detachment
Collagenase Guide
Cytochrome C
Enzyme & Inhibitor Guide
Lysing Enzymes
Nucleases
Plasma and Blood Protein
Protease Finder
Proteolytic Enzymes
Protease Inhibitors
Protein Kinase Explorer
Lipids
Microparticles
Peptide Explorer
Pharmacopoeia
PolyAmino Acids
BioFiles
Caspase
Inhibitors:
Biotin-Val-Ala-Asp(OMe)-Fluoromethyl ketone, B2057
N-T-Boc-Asp(OMe)-fluoromethyl ketone, B2682
Z-Ala-Glu(OMe)-Val-Asp(OMe)-Fluoromethyl ketone, C8484
Z-Val-Lys(Biotin)-Asp(OMe)-Fluoromethyl ketone, C8609
back to top
back to Proteolytic Enzymes Guide
Caspase 1
Product Name
Product # 
Caspase 1 human C5482
Specificity:
Release of interleukin 1b by specific cleavage at -Asp116-Ala- and -Asp27-Gly- bonds in precursor
Synthethic Substrates:
N-Acetyl-Tyr-Val-Ala-Asp p-nitroanilide, A3831
N-Acetyl-Trp-Glu-His-Asp-p-nitroanilide, A6845
N-Acetyl-Trp-Glu-His-Asp-7-amido-4-methylcoumarin, A0216
N-Acetyl-Tyr-Val-Ala-Asp-7-amido-4-methylcoumarin, A2452
N-Acetyl-Trp-Glu-His-Asp-7-amide-4-trifluoromethylcoumarin, A6720
N-Acetyl-Tyr-Val-Ala-Asp-7-amido-4-trifluoromethylcoumarin, A9965
7-Methoxycoumarin-4-acetyl-Tyr-Val-Ala-Asp-Ala-Pro-(2,4-dinitrophenyl)Lys, M5295
Inhibitors:
N-Acetyl-Trp-Glu-His-Asp-al, A1466
N-Acetyl-Tyr-Val-Ala-Asp chloromethyl ketone, A4211
N-Acetyl-Tyr-Val-Lys(biotinyl)-Asp 2,6-dimethylbenzoyloxymethyl ketone, A4339
Z-Leu-Glu-(OMe)-Val-Asp(OMe)-Fluoromethyl ketone, C9484
back to top
back to Proteolytic Enzymes Guide
Caspase 2
Product Name
Product # 
Caspase 2 human C2854
Synthethic Substrates:
N-Acetyl-Val-Asp-Val-Ala-Asp-p-nitroanilide, A5470
N-Acetyl-Val-Asp-Val-Ala-Asp-7-amido-4-trifluoromethylcoumarin, A5345
Inhibitors:
N-Acetyl-Val-Asp-Val-Ala-Asp-CHO, A2222
N-Cbz-Val-Asp(OMe)-Val-Ala-Asp(OMe)-FMK, C1605
back to top
back to Proteolytic Enzymes Guide
Caspase 3
Product Name
Product # 
Caspase 3 human C1224
Specificity:
Cleaves PARP, ICAD, gelsolin, procaspases 6 and 9
Synthethic Substrates:
N-Acetyl-Asp-Glu-Val-Asp p-nitroanilide, A2559
N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-trifluoromethylcoumarin, A0466
Acetyl-Asp-Glu-Val-Asp-7-amido-4-methylcoumarin, A1086
N-Acetyl-Asp-Gln-Met-Asp-7-amido-4-trifluoromethylcoumarin, A5595
Chromofluor 96, C7605
DFF45/ICAD human, D3815
Inhibitors:
N-AcetylL-Asp-Glu-Val-Asp-al, A0835
N-Acetyl-Ile-Glu-Thr-Asp-al, A1216
N-Acetyl-Glu-Ser-Met-Asp-al, A1341
Biotin-Asp-(OMe)-Glu-(OMe)Val(OMe)-Asp(OMe)-Fluoromethyl ketone, B1807
N-Cbz-Asp(OMe)-Gln-Met-Asp(OMe)-FMK, C0480
Caspase-4 Inhibitor (aldehyde), C0859
Z-VAD-FMK, V116
back to top
back to Proteolytic Enzymes Guide
Caspase 4
Product Name
Product # 
Caspase 4 human C6357
Synthethic Substrates:
N-Acetyl-Tyr-Val-Ala-Asp p-nitroanilide, A3831
N-Acetyl-Leu-Glu-Val-Asp-7-amido-4-trifluoromethylcoumarin, A2099
N-Acetyl-Tyr-Val-Ala-Asp-7-amido-4-methylcoumarin, A2452
N-Acetyl-Tyr-Val-Ala-Asp-7-amido-4-trifluoromethylcoumarin, A9965
back to top
back to Proteolytic Enzymes Guide
Caspase 5
Product Name
Product # 
Caspase 5 human C6482
Synthethic Substrates:
N-Acetyl-Trp-Glu-His-Asp-p-nitroanilide, A6845
N-Acetyl-Trp-Glu-His-Asp-7-amide-4-trifluoromethylcoumarin, A6720
Inhibitors:
N-Acetyl-Trp-Glu-His-Asp-al, A1466
back to top
back to Proteolytic Enzymes Guide
Caspase 6
Product Name
Product # 
Caspase 6 human C4977
Synthethic Substrates:
N-Acetyl-Val-Glu-Ile-Asp-p-nitroanilide, A5220
N-Acetyl-Val-Glu-Ile-Asp-7-amido-4-methylcoumarin, A0341
N-Acetyl-Val-Glu-His-Asp-7-amido-4-trifluoromethylcoumarin, A2097
N-Acetyl-Val-Glu-Ile-Asp-7-amido-4-trifluoromethylcoumarin, A5095
Caspase 6 Assay Kit, Fluorimetric, CASP-6-F
Inhibitors:
N-Acetyl-Val-Glu-Ile-Asp-al, A6339
Biotin-Asp-(OMe)-Glu-(OMe)Val(OMe)-Asp(OMe)-Fluoromethyl ketone, B1807
N-Cbz-Asp(OMe)-Gln-Met-Asp(OMe)-FMK, C0480
N-CBZ-Val-Glu-(OMe)-Ile-Asp(OMe)-FMK, C1730
back to top
back to Proteolytic Enzymes Guide
Caspase 7
Product Name
Product # 
Caspase 7 human C2979
Synthethic Substrates:
N-Acetyl-Asp-Glu-Val-Asp p-nitroanilide, A2559
N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-trifluoromethylcoumarin, A0466
Acetyl-Asp-Glu-Val-Asp-7-amido-4-methylcoumarin, A1086
Inhibitors:
Biotin-Asp-(OMe)-Glu-(OMe)Val(OMe)-Asp(OMe)-Fluoromethyl ketone, B1807
N-Cbz-Asp(OMe)-GLu(OMe)-Val-Asp(OMe)-FMK, C0605
back to top
back to Proteolytic Enzymes Guide
Caspase 8
Specificity:
Cleaves procaspases 3,6, and 7
Synthethic Substrates:
N-Acetyl-Ile-Glu-Thr-Asp-p-nitroanilide, A9968
Caspase 8 Colorimetric Kit, CASP-8-C
N-Acetyl-Ala-Glu-Val-Asp-7-amido-4-trifluoromethylcoumarin, A0973
N-Acetyl-Ile-Glu-Thr-Asp-7-Amido-4-methylcoumarin, A4188
N-Acetyl-Leu-Glu-Thr-Asp-7-Amido-4-trifluoromethylcoumarin, A6095
Z-Ile-Glu-Thr-Asp 7-amido-4-trifluoromethylcoumarin, C5599
Chromofluor 96, C7605
Caspase 8 Fluorimetric Kit, CASP-8-F
Inhibitors:
N-Acetyl-Ile-Glu-Thr-Asp-al, A1216
Biotin-Asp-(OMe)-Glu-(OMe)Val(OMe)-Asp(OMe)-Fluoromethyl ketone, B1807
N-Cbz-Ile-Glu(OMe)-Thr-Asp(OMe), C1230
Z-Leu-Glu-(OMe)-Thr-Asp-(OMe)-Fluoromethyl ketone, C8734
back to top
back to Proteolytic Enzymes Guide
Caspase 9
Product Name
Product # 
Caspase 9 human C8726
Specificity:
Cleaves procaspases 3,6, and 7
Synthethic Substrates:
N-Acetyl-Leu-Glu-His-Asp-p-nitroaniline, A5970
N-Acetyl-Val-Glu-His-Asp-7-amido-4-trifluoromethylcoumarin trifluoroacetate salt, A2097
N-Acetyl-Leu-Glu-His-Asp-7-amido-4-trifluoromethylcoumarin, A5845
Chromofluor 96, C7605
Inhibitors:
N-Cbz-Leu-Glu(OMe)-His-Asp(OMe)-FMK, C1355
back to top
back to Proteolytic Enzymes Guide
Caspase 10
Product Name
Product # 
Caspase 10 human, recombinant, expressed in Escherichia coli >90% (SDS-PAGE) Lyophilized powder 8,000 units/mg protein C7982
Caspase 10 human, recombinant, expressed in Escherichia coli >90% (SDS-PAGE) Lyophilized powder 8,000 units/mg protein (Bradford) C6607
Specificity:
Cleaves procaspases 3, 4, 6, 7, 8 and 9
Inhibitors:
Biotin-Asp-(OMe)-Glu-(OMe)Val(OMe)-Asp(OMe)-Fluoromethyl ketone, B1807
back to top
back to Proteolytic Enzymes Guide
Caspase 13
Inhibitors:
Z-Leu-Glu(OMe)-Glu(OMe)-Asp(OMe)-Fluoromethyl ketone, C8859
back to top
back to Proteolytic Enzymes Guide
Cathepsin B, E.C. 3.4.22.1
Product Name
Product # 
Cathepsin B from bovine spleen C6286
Cathepsin B from human liver C8571
Cathepsin B from human placenta C0150
Specificity:
Hydrolysis of proteins with broad specificity for peptide bonds. Preferentially cleaves -Arg-Arg bonds in small molecule substrates (thus differing from cathepsin L). In addition to being an endopeptidase, shows peptidyl-dipeptidase activity, liberating C-terminal dipeptides
Synthethic Substrates:
Na-Benzoyl-L-arginine 4-methoxy-beta-naphthylamide hydrochloride, B0885
Z-Val-Lys-Lys-Arg-4-methoxy-beta-naphthylamide trihydrochloride, C2772
Z-Phe-Arg 4-methoxy-beta-naphthylamide hydrochloride, C3282
Z-Arg-Arg-7-amido-4-Z-RR-AMC-methylcoumarin hydrochloride, C5429
Z-Arg-Arg 4-methoxy-beta-naphthylamide acetate salt, C5520
Z-Ala-Arg-Arg 4-methoxy-beta-naphthylamide acetate salt, C8536
N-Succinyl-Leu-Leu-Val-Tyr-7-Amido-4-Methylcoumarin, S6510
Hippuryl-Arg, H2508
Inhibitors:
N-Acetyl-Leu-Leu-Met-al, A6060
N-Acetyl-Leu-Leu-Norleu-al, A6185
Antipain, A6191
Biotin-Phe-Ala-Fluoromethyl ketone, B1932
Cystatin, C0408
CA-074, C5732
CA-074 Methyl ester, C5857
Chymostatin, C7268
Cystatin, C8917
Z-Phe-Phe-Fluoromethyl ketone, C9109
E-64, E3132
Leupeptin, L2023
MeOSuc-Phe-homo-Phe fluoromethyl ketone, M5314
a2-Macroglobulin, M6159
Procathepsin B, rat, fragment 26-50, P0350
Procathepsin B, rat, fragment 36-50, P0475
back to top
back to Proteolytic Enzymes Guide
Cathepsin C, E.C. 3.4.14.1
Product Name
Product # 
Cathepsin C from bovine spleen C8511
Specificity:
Release of an N-terminal dipeptide, Xaa-Xbb Xcc-, except when Xaa is Arg or Lys, or Xbb or Xcc is Pro
Synthethic Substrates:
Gly-Phe p-nitroanilide, G0142
Gly-Phe beta-naphthylamide, G4255
Gly-Phe beta-naphthylamide, G9512
Ser-Tyr beta-naphthylamide, S5628
Inhibitors:
Iodoacetic Acid, I2512
back to top
back to Proteolytic Enzymes Guide
Cathepsin D, E.C. 3.4.23.5
Product Name
Product # 
Cathepsin D from bovine spleen C3138
Cathepsin D from human liver C8696
Specificity:
Specificity similar to, but narrower than, that of pepsin A. Does not cleave the Gln4-His bond in B chain of insulin
Synthethic Substrates:
Boc-Phe-Ala-Ala-p-Nitro-Phe-Phe-Val-Leu 4-hydroxymethylpyridine ester, B1644
N-Acetyl-Arg-Gly-Phe-Phe-Pro-7-amido-4-methylcoumarin, C6358
N-Acetyl-Arg-Gly-Phe-Phe-Pro-7-amido-4-trifluoromethylcoumarin, C6983
N-Acetyl-Arg-Gly-Phe-Phe-Pro 4-methoxy-2-naphthylamide, C7108
7-Methoxycoumarin-4-Acetyl-Gly-Lys-Pro-Ile-Leu-Phe-Phe-Arg-Leu-Lys(DNP)-D-Arg-amide, M0938
Inhibitors:
Antipain, A6191
Chymostatin, C7268
Pepstatin A, P5318
back to top
back to Proteolytic Enzymes Guide
Cathepsin G, E.C. 3.4.21.20
Product Name
Product # 
Cathepsin G from human leukocytes C4428
Cathepsin G from human sputum C5108
Specificity:
Specificity similar to chymotrypsin C
Synthethic Substrates:
N-Methoxysuccinyl-Ala-Ala-Pro-Met p-nitroanilide, M7771
N-Succinyl-Ala-Ala-Pro-Phe p-nitroanilide, S7388
N-Succinyl-Gly-Gly-Phe-7-amido-4-trifluoromethylcoumarin, S4689
N-Succinyl-Gly-Gly-Phe-4-methoxy-2-naphthylamide, S4814
N-Succinyl-Ala-Ala-Pro-Phe thiobenzyl ester, S6518
N-Succinyl-Phe-Leu-Phe thiobenzyl ester, S6643
Inhibitors:
a1-Antchymotrypsin, A9285
Chymostatin, C7268
Z-Gly-Leu-Phe-Chloromethyl ketone, C9984
Diisopropyl fluorophosphate, D0879
3,4-Dichloroisocoumarin, D7910
N-Acetyl-eglin C, E7888
back to top
back to Proteolytic Enzymes Guide
Cathepsin H, E.C. 3.4.22.16
Product Name
Product # 
Cathepsin H from human liver C8821
Specificity:
Hydrolysis of proteins, acting as an aminopeptidase (notably, cleaving Arg bonds) as well as an endopeptidase
Synthethic Substrates:
L-Arginine-7-amido-4-methylcoumarin hydrochloride, A2027
back to top
back to Proteolytic Enzymes Guide
Cathepsin L, E.C. 3.4.22.15
Product Name
Product # 
Cathepsin L from human liver C6854
Specificity:
Similar to that of papain. As compared to cathepsin B, cathepsin L exhibits higher activity towards protein substrates and no peptidyl-dipeptidase activity
Synthethic Substrates:
Z-Phe-Arg 4-methoxy-beta-naphthylamide hydrochloride, C3282
Inhibitors:
N-Acetyl-Leu-Leu-Norleu-al, A6185
back to top
back to Proteolytic Enzymes Guide
Chymopapain, E.C. 3.4.22.6
Product Name
Product # 
Chymopapain from papaya latex C8526
Specificity:
Similar to that of papain
Synthethic Substrates:
BAEE, B4500
back to top
back to Proteolytic Enzymes Guide