Sigma-Aldrich
Biochemicals
Antibiotic Explorer
Biological Buffers
Biological Detergents
Books
Enzyme Explorer
Application Index
Class Index
Inhibitor Index
Substrate Index
Cofactor Index
Lectin Index
Cell Signaling Enzymes
Analytical Enzymes
Enzyme Reagents
Key Resources
Enzyme Literature
Assay Library
Carbohydrate Analysis
Collagenase Guide
Cytochrome C
Enzyme & Inhibitor Guide
Lysing Enzymes
Nucleases
Plasma and Blood Protein
Protease Finder
Proteolytic Enzymes
Protease Inhibitors
Protein Kinase Explorer
Lipids
Peptide Explorer
Pharmacopoeia
PolyAmino Acids
Radiochemicals
Vitamins and Derivatives
 "; } else { print "
"; } ?>
Chymase, E.C. 3.4.21.39
Product Name
Product # 
Chymase C9612
Specificity:
Preferential cleavage: Phe-Xaa > Tyr-Xaa > Trp-Xaa > Leu-Xaa
Synthethic Substrates:
BTEE, B6125
back to top
back to Proteolytic Enzymes Guide
Chymotrypsin, a-, E.C. 3.4.21.1
Product Name
Product # 
a-Chymotrypsin from bovine pancreas, suitable for protein sequencing Salt-free, lyophilized powder C6423
b-Chymotrypsin from bovine pancreas C4629
a-Chymotrypsin Insoluble enzyme attached to carboxymethyl cellulose from bovine pancreas C7260
a-Chymotrypsin-Agarose from bovine pancreas C9134
a-Chymotrypsin DFP treated C1012
a-Chymotrypsin from human pancreas, Lyophilized powder C8946
a-Chymotrypsin from bovine pancreas, Type II Essentially salt-free, lyophilized powder >40 units/mg protein C4129
a-Chymotrypsin from bovine pancreas, Type I-S Essentially salt-free, lyophilized powder 40-60 units/mg protein C7762
a-Chymotrypsin TLCK Treated from bovine pancreas C3142
Specificity:
Preferential cleavage: Ty-Xaa, Trp-Xaa, Phe-Xaa, Leu-Xaa
Synthethic Substrates:
N-Succinyl-Ala-Ala-Pro-Phe p-nitroanilide, S7388
N-Succinyl-Ala-Ala-Pro-Phe-7-amido-4-methylcoumarin, S9761
Ala-Ala-Phe-7-amido-4-methylcoumarin, A3401
ALA-ALA-VAL-ALA P-Nitroanilide, A9273
BTEE, B6125
N-Benzoyl-L-tyrosine p-nitroanilide , B6760
Glutaryl-Ala-Ala-Phe-4-methoxy-b-naphthylamide, G3769
N-Glutaryl-Gly-Gly-Phe-b-naphthylamide, G4636
4-Methylumbelliferyl p-trimethylammoniocinnamate chloride, M4507
N-Succinyl-Gly-Gly-Phe p-nitroanilide, S1899
N-Succinyl-L-phenylalanine p-nitroanilide, S2628
N-Succinyl-Ala-Ala-Pro-Phe 7-amido-4-trifluoromethylcoumarin, S5189
N-Succinyl-Ala-Ala-Pro-Phe 4-methoxy-2-naphthylamide, S5314
Inhibitors:
Z-L-Phe chloromethyl ketone, C9511
a2-Antiplasmin, A0914
Aprotinin, A1153
e-Amino-n-caproic acid, A2504
a1-Antitrypsin, A6150
e-Amino-n-caproic acid, A7824
4-(2-Aminoethyl) benzenesulfonyl fluoride hydrochloride, A8456
a1-Antitrypsin, A9024
a1-Antchymotrypsin, A9285
Bromoenol lactone, B1552
Chymostatin, C7268
Diisopropyl fluorophosphate, D0879
Ecotin, E1398
N-Acetyl-eglin C, E7888
Gabexate mesylate, G2417
Leupeptin, L2023
Trypsin Inhibitor, T0256
Trypsin Inhibitor, T1886
N-Tosyl-L-phenylalanine chloromethyl ketone, T4376
Trypsin Inhibitor, T4385
Trypsin Inhibitor, T9253
Trypsin Inhibitor, T9378
Trypsin-chymotrypsin inhibitor, T9777
back to top
back to Proteolytic Enzymes Guide
Clostripain, E.C. 3.4.22.8
Product Name
Product # 
Clostripain from Clostridium histolyticum C0888
Specificity:
Preferential cleavage: Arg-Xaa , including Arg-Pro, but not Lys-
Synthethic Substrates:
BAEE, B4500
Inhibitors:
p-Chloromercuribenzoic acid Free Acid, C5913
Histatin 5, H6027
N a-p-Tosyl-L-lysine chloromethyl ketone hydrochloride, T7254
back to top
back to Proteolytic Enzymes Guide
Collagenase, E.C. 3.4.24.3
Product Name
Product # 
Collagenase from Clostridium histolyticum, 2-5 FALGPA units/mg solid >1200 CDU/mg solid (CDU = collagen digestion units) Type XI C7657
Collagenase from Clostridium histolyticumType VII powder 1000-3000 CDU/mg solid (CDU = collagen digestion units) 4-12 FALGPA units/mg protein cell culture, tested C2799
Collagenase from Clostridium histolyticum, cell culture, tested Lyophilized powder Type XI-S C9697
Collagenase from Clostridium histolyticum, Type XI powder >1200 CDU/mg solid (CDU = collagen digestion units) 2-5 FALGPA units/mg protein cell culture, tested C9407
Collagenase/Dispase from Achromobacter iophagus/Bacillus polymyxa C3180
Collagenase from Clostridium histolyticum, Type I-A powder >125 CDU/mg solid (CDU = collagen digestion units) 0.5-2.0 FALGPA units/mg protein cell culture, tested C2674
Collagenase from Clostridium histolyticum, Type VII-S cell culture, tested Lyophilized powder C9572
Collagenase from Clostridium histolyticum, Type III Fraction A Lyophilized powder 2-10 FALGPA units/mg protein minimum 400 CDU/mg solid (CDU = collagen digestion units) C0255
Collagenase from Clostridium histolyticum, Type VII Lyophilized powder 1,000-3,000 CDU/mg solid (CDU = collagen digestion units) 4-12 FALGPA units/mg protein C0773
Collagenase from Clostridium histolyticum, 1.1-1.5 FALGPA units/mg protein (Neutral Protease: 20-50units per mg solid) Sigma Blend Type H C8051
Collagenase from Clostridium histolyticum, 1-3 FALGPA units/mg protein >125 CDU/mg solid (CDU = collagen digestion units) Type V C9263
Collagenase from Clostridium histolyticum, 1.8-2.2 FALGPA units/mg protein (Neutral Protease:<10 units per mg solid) Sigma Blend Type F C7926
Collagenase from Clostridium histolyticum, 0.5-0.9 FALGPA units/mg protein (Neutral Protease: 50-80 units per mg solid) Sigma Blend Type L C8176
Collagenase + protease inhibitor, suitable (Each lot is evaluated for suitability in the release of physiologically active rat pancreatic islets (for methodology see Type V)) 2-5 FALGPA units/mg protein minimum C6079
Collagenase from Clostridium histolyticum, Essentially salt-free, lyophilized powder 0.5-3.0 FALGPA units/mg protein >125 CDU/mg solid (CDU = collagen digestion units) Type IA C9891
Collagenase from Clostridium histolyticum, Essentially salt-free, lyophilized powder 0.25-1.0 FALGPA units/mg protein >125 CDU/mg solid (CDU = collagen digestion units) Type I C0130
Collagenase from Clostridium histolyticum, suitable for release of rat epididymal adipocytes and hepatocytes (for methodology see Type II and Type IV) 0.5-3.0 FALGPA units/mg solid >125 CDU/mg solid (CDU = collagen digestion units) Type VIII C2139
Collagenase from Clostridium histolyticum, 0.5-3.0 FALGPA units/mg solid >125 CDU/mg solid (CDU = collagen digestion units) Type II C6885
Collagenase from Clostridium histolyticum, 0.5-3.0 FALGPA units/mg solid >125 CDU/mg solid (CDU = collagen digestion units) Type IV C5138
Collagenase from Clostridium histolyticum, Type I-AS cell culture, tested Lyophilized powder C9722
Specificity:
Digestion of native collagen in the triple helical region at Gly bonds. With synthetic peptides, a preference is shown for Gly at P3 and P1'; Pro and Ala at P2 and P2'; and hydroxyproline, Ala or Arg at P3'
Synthethic Substrates:
Z-Gly-Pro-Leu-Gly-Pro, C0627
Collagen Type I-FITC, C4361
Collagen Type II-FITC, C4486
Collagen-Fluorescein, C5858
Collagen-Fluorescein, C8008
N-(2,4-Dinitrophenyl)-Pro-Leu-Gly-Cys(Me)-His-Ala-D-Arg amide, D0296
FALGPA, F5135
Inhibitors:
5-(R,S)-T-trans-Cinnamido-7-methyl-4-oxo-octanoyl-L-prolyl-L-proline, C0360
Dichloromethylenediphosphonic acid disodium salt, D4434
Ecotin, E1398
EDTA, ED2SS
Isoamylphosphonyl-Gly-L-Pro-L-Ala, dipotassium, I7029
(2R)-2-Mercaptomethyl-4-methylpentanoyl-beta-(2-naphthyl)-Ala-Ala Amide, M3531
back to top
back to Proteolytic Enzymes Guide
Complement C1r, E.C. 3.4.21.41
Product Name
Product # 
Complement C1r from human serum C2162
Specificity:
Selective cleavage of Lys(or Arg) Ile bond in complement subcomponent C1s
Inhibitors:
Complement C1 esterase inhibitor, C2412
Diisopropyl fluorophosphate, D0879
C1 Esterase inhibitor, E0518
back to top
back to Proteolytic Enzymes Guide
Complement C1s, E.C. 3.4.21.42
Specificity:
Cleavage of Arg-Ala bond in complement component C4 to form C4a and C4b, and Lys(or Arg) Lys bond in complement component C2 to form C2a and C2b: the "classical" pathway C3 convertase
Inhibitors:
Complement C1 esterase inhibitor, C2412
Diisopropyl fluorophosphate, D0879
C1 Esterase inhibitor, E0518
back to top
back to Proteolytic Enzymes Guide
Complement Factor D, E.C. 3.4.21.46
Product Name
Product # 
Complement factor D from human plasma C5688
Specificity:
Selective cleavage of Arg-Lys bond in complement factor B when in complex with complement subcomponent C3b or with cobra venom factor
Inhibitors:
Diisopropyl fluorophosphate, D0879
back to top
back to Proteolytic Enzymes Guide
Complement factor I, E.C. 3.4.21.45
Product Name
Product # 
Complement factor I from human plasma C5938
Specificity:
Inactivates complement subcomponents C3b, iC3b and C4b by proteolytic cleavage
back to top
back to Proteolytic Enzymes Guide
Cucumisin, E.C. 3.4.21.25
Product Name
Product # 
Cucumisin C1932
Specificity:
Hydrolysis of proteins with broad specificity
back to top
back to Proteolytic Enzymes Guide
Dipeptidyl peptidase IV, E.C. 3.4.14.5
Product Name
Product # 
Dipeptidyl Peptidase from porcine kidney D7052
Specificity:
Release of an N-terminal dipeptide, Xaa-Xbb Xcc-, from a polypeptide, preferentially when Xbb is Pro, provided Xcc is neither Pro nor hydroxyproline
Synthethic Substrates:
Gly-Pro-7-amido-4-methylcoumarin hydrobromide, G2761
Gly-Pro p-nitroanilide hydrochloride, G0513
Inhibitors:
Ile-Pro-Ile, I9759
back to top
back to Proteolytic Enzymes Guide
Elastase, leukocyte, E.C. 3.4.21.37
Product Name
Product # 
Elastase from human leukocytes E8140
Specificity:
Hydrolysis of proteins, including elastin. Preferential cleavage Val-Xaa > Ala-Xaa
Synthethic Substrates:
Boc-Ala-Pro-Nva p-chlorothiobenzyl ester, B2017
N-Methoxysuccinyl-Ala-Ala-Pro-Val-7-amido-4-trifluoromethylcoumar, M4563
N-(Methoxysuccinyl)-Ala-Ala-Pro-Val 4-nitroanilide, M4765
N-Methoxysuccinyl-Ala-Ala-Pro-Val-7-amido-4-methylcoumarin, M9771
Inhibitors:
Aprotinin, A1153
Diisopropyl fluorophosphate, D0879
3,4-Dichloroisocoumarin, D7910
Eglin C fragment 60-63 methyl ester, E2017
N-Acetyl-eglin C, E7888
a2-Macroglobulin, M6159
Phenylmethylsulfonyl fluoride, P7626
back to top
back to Proteolytic Enzymes Guide
Elastase, pancreatic, E.C. 3.4.21.36
Product Name
Product # 
Elastase, pancreatic solution from porcine pancreas, Type I Aqueous suspension minimum 4 units/mg protein E1250
Elastase, pancreatic from porcine pancreas, Lyophilized powder 3-6 units/mg protein (biuret) cell culture, tested E7885
Elastase, pancreatic from porcine pancreas, Type IV Lyophilized powder minimum 4 units/mg protein (biuret) E0258
Elastase, pancreatic from porcine pancreas, Type II-A Lyophilized powder minimum 1 units/mg protein (biuret) E6883
Elastase, pancreatic from porcine pancreas, Type III Lyophilized powder minimum 4 units/mg protein E0127
Specificity:
Hydrolysis of proteins, including elastin. Preferential cleavage: Ala-Xaa
Synthethic Substrates:
N-Methoxysuccinyl-Ala-Ala-Pro-Val-7-amido-4-methylcoumarin, M9771
Inhibitors:
a1-Antitrypsin, A6150
a1-Antitrypsin, A9024
Diisopropyl fluorophosphate, D0879
3,4-Dichloroisocoumarin, D7910
Ecotin, E1398
a2-Macroglobulin, M6159
back to top
back to Proteolytic Enzymes Guide
Endoproteinase Arg-C, E.C. 3.4.21.35
Product Name
Product # 
Endoproteinase Arg-C murine submaxillary gland, suitable for protein sequencing Lyophilized powder P6056
Endoproteinase Arg-C murine submaxillary gland, Type XX-S Lyophilized powder 250-800 units/mg protein (Lowry) P5171
Specificity:
Cleaves specifically at the carboxyl side of arginine residues. The enzyme has also been shown to cleave Lys-Lys and Lys-Arg bonds; all Arg-X bonds may not be hydrolyzed. This enzyme also exhibits esterase and amidase activities.
Synthethic Substrates:
TAME, T4626
Inhibitors:
Diisopropyl fluorophosphate, D0879
a2-Macroglobulin, M6159
N-a-p-Tosyl-L-lysine chloromethyl ketone Hydrochloride, T7254
back to top
back to Proteolytic Enzymes Guide
Endoproteinase Asp-N, E.C. 3.4.24.33
Product Name
Product # 
Endoproteinase Asp-N from Pseudomonas fragi mutant strain P3303
Specificity:
Cleavage of Xaa-Asp, Xaa-Glu and Xaa-cysteic acid bonds
back to top
back to Proteolytic Enzymes Guide
Endoproteinase Glu-C, E.C. 3.4.21.19
Product Name
Product # 
Endoproteinase Glu-C Staphylococcus aureus V8, suitable for protein sequencing Lyophilized powder P6181
Endoproteinase Glu-C Staphylococcus aureus V8, Type XVII-B Lyophilized powder 500-1,000 units/mg solid P2922
Specificity:
Preferential cleavage: Glu-Xaa, Asp-Xaa
Inhibitors:
Diisopropyl fluorophosphate, D0879
a2-Macroglobulin, M6159
back to top
back to Proteolytic Enzymes Guide
Endoproteinase Lys-C, E.C. 3.4.21.50
Product Name
Product # 
Endoproteinase Lys-C from Lysobacter enzymogenes, suitable for protein sequencing Lyophilized powder P3428
Endoproteinase Lys-C from Lysobacter enzymogenes, Lyophilized powder P2289
Specificity:
Cleaves at the carboxyl side of lysyl residues. Lys-Pro and Lys-Glu bonds are also cleaved
Synthethic Substrates:
N-(p-Tosyl)-Gly-Pro-Lys 4-nitroanilide acetate salt, T6140
Inhibitors:
Aprotinin, A1153
Leupeptin, L2023
Diisopropyl fluorophosphate, D0879
N-a-p-Tosyl-L-lysine chloromethyl ketone Hydrochloride, T7254
back to top
back to Proteolytic Enzymes Guide
Enterokinase, E.C. 3.4.21.9
Product Name
Product # 
Enterokinase from bovine intestine, powder E5144
Enterokinase porcine intestine, Lyophilized powder 100-500 units/mg protein E0885
Enterokinase from bovine intestine, Salt-free, lyophilized powder >0.5 units/mg solid E5510
Enterokinase porcine intestine, Salt-free, lyophilized powder approx. 2 units/mg solid E0632
Enterokinase solution from calf intestine, Buffered aqueous glycerol solution minimum 1,000 units/mg protein (BCA) E0766
Specificity:
Activation of trypsinogen by selective cleavage of Lys6-Ile bond
Synthethic Substrates:
Gly-Asp-Asp-Asp-Asp-Lys-b-naphthylamide, G5261
Inhibitors:
Diisopropyl fluorophosphate, D0879
a2-Macroglobulin, M6159
back to top
back to Proteolytic Enzymes Guide
Factor Xa, E.C. 3.4.21.6
Product Name
Product # 
Factor X Activated (Xa) bovine plasma F2027
Specificity:
Selective cleavage of Arg-Thr and then Arg-Ile bonds in prothrombin to form thrombin
Synthethic Substrates:
N-Benzoyl-Ile-Glu-Gly-Arg p-nitroanilide hydrochloride, B2291
N-Benzoyl-Ile-Glu-Gly-Arg p-nitroanilide acetate salt, B7020
Boc-Ile-Glu-Gly-Arg-7-amido-4-methylcoumarin hydrochloride, B9760
Factor Xa chromogenic substrate, F3301
4-Methylumbelliferyl 4-guanidinobenzoate hydrochloride monohydrate, M4149
Inhibitors:
a2-Antiplasmin, A0914
Antithrombin III, Human, A2221
Antithrombin III, Rat, A6397
Antithrombin III, Bovine, A9141
Diisopropyl fluorophosphate, D0879
3,4-Dichloroisocoumarin, D7910
Ecotin, E1398
Phenylmethylsulfonyl fluoride, P7626
Trypsin Inhibitor, T9003
back to top
back to Proteolytic Enzymes Guide