|
Lectin Index |
 |
Enzyme Explorer |
 |
 |
| |
|
Sigma has the largest selection of lectins and lectin conjugates available for the life scientist. Over 100 products from over 35 sources make Sigma the single best source for your lectin requirements.
Features and Benefits:
Backed by quality and service youve come to expect, Sigma offers lectins for all your life science applications, including;
- Histochemical Studies
- Lymphocyte Subpopulations Studies
- Cell Fractionation
|
- Mitogenic Stimulation
- Blood Group Typing
|
Link To:
Taxonomic Source Listing
Conjugate Details
Product List (includes conjugates, specificity, mitogenic
activity, and more)
Taxonomic Source Listing:
| Common
Name |
Taxonomic
Name |
|
Common
Name |
Taxonomic
Name |
| *Abrin |
Abrus precatorius |
Jimson weed |
Datura stramonium |
| Asparagus pea |
Tetragonolobus
purpureas |
*Labumum, Scotch |
Labumum alpinum |
| *Avocado |
Perseau americana |
Lentil |
Lens culinaris |
| *Bitter pear melon |
Momordica charantia |
Lotus |
Tetragonolobus
purpureas |
| Broad bean |
Vicia faba |
Mistletoe, European |
Viscum album |
| Camels foot tree |
Bauhinia purpurea |
*Mung bean |
Vigna radiata |
| Castor bean |
Ricinus communis |
Mushroom |
Agaricus bisporus |
| Chick pea |
Cicer arietinum |
Osage orange |
Maclura pomifera |
| *Cobra, Mozambique |
Naja mocambique
mocambique |
*Pagoda tree |
Sophora japonica |
| *Cobra, Thailand |
Naja naja kaouthia |
Pea, garden |
Pisum sativum |
| Con A |
Canavalia ensiformis |
Peanut |
Arachis hykpogaea |
| Concanavalin A |
Canavalia ensiformis |
Pokeweek |
Phytolacca americana |
| Coral tree |
Erythrina cristagalli |
Potato |
Solanum tuberosum |
| Coral tree, Israel |
Erythrina corallodendron |
Red kidney bean |
Phaseolus vulgaris |
| Daffodil |
Narcissus pseudonarcissus |
*Red marine algae |
Ptilota plumosa |
| Eel |
Anguilla anguilla |
Roman snail |
Helix pomatia |
| Elder |
Sambucus nigra |
Scarlet runner bean |
Phaseolus coccineus |
| Fava bean |
Vicia faba |
Siberian pea tree |
Caragana arborescens |
| Furze |
Ulex europaeus |
Snail, garden |
Helix aspersa |
| Gorse |
Ulex europaeus |
Snowdrop |
Galanthus nivalis |
| Green marine algae |
Codium fragile |
Soybean |
Glycine max |
| Hairy vetch |
Vicia villosa |
Spindle tree |
Euonymus eruopaeus |
| Horse gram |
Dolichos biflorus |
*Sweet pea |
Lathyrus odoratus |
| Horseshoe crab |
Limulus polyphemus |
Thorn apple |
Datura stramonium |
| Jacalin |
Artocarpus integrifolia |
Tomato |
Lycopersicon esculentum |
| Jack bean |
Canavalia ensiformis |
Wheat germ |
Triticum vulgaris |
| Japanese wisteris |
Wisteria floribunda |
Winged bean |
Psophocarpus tetragonolobus |
| *Jequity bean |
Abrus precatorius |
Winged pea |
Tetragonolobus
purpureas |
| *Not currently available, please
click here to inquire. |
Top of Page |
Conjugate Details:
Sigma has the largest selection of
lectins and lectin conjugates available for the life scientist.
Over 100 products from over 35 natural sources. Conjugates include:
Agarose FITC Peroxidase
Biotin TRITC Gold
Many lectins are available as conjugates. Conjugation does not alter the specificity of the lectin. The degree of conjugation is expressed as 1) a mole dye per mole lectin basis 2) a unit enzyme activity per mg protein or 3) a mg protein per ml packed gel basis. For colloidal gold conjugates, the activity of the lectin is determined by a dot blot assay using an appropriate glycoprotein. Agglutination activity is expressed in microgram per ml and is determined from serial dilution studies of a 1 mg per ml solution. This activity is the lowest concentration of lectin to agglutinate a 2% suspension of appropriate erythrocytes after
1 hour incubation at 25 0C.
Top of Page
Lectins Product List:
|
Prod.
No.: |
LECTIN: |
Mol. Wt.
(kDa): |
Subunits: |
Specificity
Blood
Group: |
Specificity
Sugar: |
Mitogenic
Activity: |
Conjugates: |
| L
5640 |
Agaricus bisporus |
58.5 |
|
|
ß-gal(1->3)galNAc |
|
|
| L
4141 |
Anguilla anguilla |
40 |
2 |
H |
a-L-Fuc |
|
|
| L
0881 |
Arachis hypogaea |
120 |
4 |
T |
ß-gal(1->3)galNAc |
|
|
| L
7759 |
conjugate |
|
|
Peroxidase |
| L
6135 |
conjugate |
|
|
Biotin |
| L
7381 |
conjugate |
|
|
FITC |
| L
3766 |
conjugate |
|
|
TRITC |
| L
6646 |
conjugate |
|
|
Agarose |
| L
3515 |
Artocarpus integrifolia |
42 |
4 |
T |
a-gal->OMe |
(+) |
|
| L
4650 |
conjugate |
|
|
Peroxidase |
| L
5147 |
conjugate |
|
|
Agarose |
| Bandeiraea
Simplicifolia |
Top
of Page |
| L
2380 |
BS-I |
114 |
4 |
A, B |
a-gal,
a-galNAc |
|
|
| L
3759 |
conjugate |
|
|
Biotin |
| L
9381 |
conjugate |
|
|
FITC |
| L
5264 |
conjugate |
|
|
TRITC |
| L
1509 |
BS-I-A4 |
114 |
4 |
A |
a-galNAc |
|
|
| L
0890 |
conjugate |
|
|
FITC |
| L
3019 |
BS-I-B4 |
114 |
4 |
B |
a-gal |
|
|
| L
5391 |
conjugate |
|
|
Peroxidase |
| L
2140 |
conjugate |
|
|
Biotin |
| L
2895 |
conjugate |
|
|
FITC |
| L
6013 |
Bauhinia purpurea |
195 |
4 |
|
ß-gal(1->3)galNAc |
(+) |
|
| L
9637 |
Caragana arborescens |
60;120 (c) |
2;4 |
|
galNAc |
|
|
| L
3141 |
Cicer arietinum |
44 |
2 |
|
fetuin |
|
|
| L
2638 |
Codium fragile |
60 |
4 |
|
galNAc |
|
|
| L
7275 |
Concanavalin A |
102 |
4 |
|
a-man,
a-glc |
(+) |
|
| L
6397 |
conjugate |
|
|
Peroxidase |
| C
2272 |
conjugate |
|
|
Biotin |
| C
7642 |
conjugate |
|
|
FITC |
| C
6904 |
conjugate |
|
|
Agarose |
| C
9017 |
conjugate |
|
|
Sepharose |
| L
5021 |
conjugate |
|
|
Gold, 10nm |
| L
3642 |
conjugate |
|
|
Gold, 20nm |
| C
7898 |
conjugate |
|
|
Ferritin |
| L
3885 |
Succinyl-Concanavalin A |
51 |
2 |
|
a-man,
a-glc |
(+) (d) |
|
| L
9385 |
conjugate |
|
|
FITC |
| L
2766 |
Datura stramonium |
86 |
2(a&ß)a |
|
(glcNAc)2 |
|
|
| L
2785 |
Dolichos biflorus |
140 |
4 |
A1 |
a-galNAc |
|
|
| L
1287 |
conjugate |
|
|
Peroxidase |
| L
6533 |
conjugate |
|
|
Biotin |
| L
9142 |
conjugate |
|
|
FITC |
| L
9658 |
conjugate |
|
|
TRITC |
| L
9894 |
conjugate |
|
|
Agarose |
| L
2142 |
Erythrina corallodendron |
60 |
2 |
|
ß-gal(1->4)glcNAc |
(+) |
|
| L
5390 |
Erythrina cristagalli |
56.8 |
2(a&ß)a |
|
ß-gal(1->4)glcNAc |
|
|
| L
7400 |
Euonymus europaeus |
166 |
4(a&ß)a |
B, H |
a-gal(1->3)gal |
(+) |
|
| L
8725 |
Galanthus nivalis |
52 |
4 |
(h) |
non-reduc. D-man |
|
|
| L
8775 |
conjugate |
|
|
Agarose |
| L
1395 |
Glycine max |
110 |
4 |
|
galNAc |
(+) (b) |
|
| L
2650 |
conjugate |
|
|
Peroxidase |
| L
4511 |
conjugate |
|
|
TRITC |
| L
1105 |
conjugate |
|
|
Agarose |
| L
6635 |
Helix aspersa |
79 |
|
A |
galNAc |
|
|
| L
8764 |
conjugate |
|
|
Biotin |
| L
3382 |
Helix pomatia |
79 |
6 |
A |
galNAc |
|
|
| L
6387 |
conjugate |
|
|
Peroxidase |
| L
6512 |
conjugate |
|
|
Biotin |
| L
1034 |
conjugate |
|
|
FITC |
| L
1261 |
conjugate |
|
|
TRITC |
| L
9267 |
Lens culinaris |
49 |
2 |
|
a-man |
(+) |
|
| L
4143 |
conjugate |
|
|
Biotin |
| L
9262 |
conjugate |
|
|
FITC |
| L
0511 |
conjugate |
|
|
Sepharose |
| L
2263 |
Limulus polyphemus |
400 |
18 |
|
NeuNAc |
|
|
| L
2886 |
Lycopersicon esculentum |
71 |
|
|
(glcNAc)3 |
(+) (e) |
|
| L
0651 |
conjugate |
|
|
Biotin |
| L
0401 |
conjugate |
|
|
FITC |
| L
8025 |
Maackia amurensis |
130 |
2(a&ß) |
O |
sialic acid |
(+) |
|
| L
6141 |
Maclura pomifera |
40-43 |
2(a&ß)a |
|
a-gal,a-galNAc |
|
|
| L
4401 |
conjugate |
|
|
Peroxidase |
| L
2013 |
conjugate |
|
|
Biotin |
| L
5650 |
Narcissus pseudonarcissus |
26 |
2 |
(h) |
a-D-man |
|
|
| L
3138 |
Phaseolus coccineus |
112 |
4 |
|
| L
4389 |
conjugate |
|
|
TRITC |
| Phaseolus
Vulgaris |
Top
of Page |
| L
8629 |
PHA-E |
128 |
4 |
|
oligosaccharide |
(+) |
|
| L
6139 |
conjugate |
|
|
TRITC |
| L
2769 |
PHA-L |
126 |
4 |
|
oligosaccharide |
(+) |
|
| L
8754 |
PHA-P |
|
| L
2646 |
PHA-M |
|
| L
9379 |
Phytolacca americana |
32f |
|
|
(glcNAc)3 |
(+) |
|
| L
2882 |
conjugate |
|
|
Agarose |
| L
5380 |
Pisum sativum |
49 |
4(a&ß)a |
|
a-man |
(+) |
|
| L
0770 |
conjugate |
|
|
FITC |
| L
9895 |
Pseudomonas aeruginosa (PA-I) |
13-13.7 |
|
|
gal |
|
|
| L
2138 |
Psophocarpus tetragonolobus |
35 |
1 |
|
galNAc, gal |
|
|
| L
3139 |
conjugate |
|
|
Peroxidase |
| L
3014 |
conjugate |
|
|
Biotin |
| L
3264 |
conjugate |
|
|
FITC |
| Ricinus
Communis |
Top
of Page |
| L
7886 |
Agglutinin, RCA120 |
120 |
4 |
|
ß-gal |
|
|
| L
2390 |
conjugate |
|
|
Agarose |
| L
2758 |
conjugate |
|
|
Peroxidase |
| L
9514 |
Ricin, A chain |
|
| L
4022 |
Ricin, A chain, deglycosylated |
|
| L
9639 |
Ricin, B chain |
|
| L
6890 |
Sambucus nigra |
|
| L
4266 |
Solanum tuberosum |
|
| L
9254 |
Tetragonolobus purpureas |
120(A), 58(B), 117(C) |
4;2;4 |
H |
a-L-fuc |
|
|
| L
5759 |
conjugate |
|
|
Peroxidase |
| L
3134 |
conjugate |
|
|
Biotin |
| L
5644 |
conjugate |
|
|
FITC |
| L
9640 |
Triticum vulgaris |
36 |
2 |
|
(glcNAc)2, NeuNAc |
(+) |
|
| L
3892 |
conjugate |
|
|
Peroxidase |
| L
0390 |
conjugate |
|
|
Peroxidase
(inactive) |
| L
5142 |
conjugate |
|
|
Biotin |
| L
4895 |
conjugate |
|
|
FITC |
| L
9884 |
conjugate |
|
|
Evans Blue |
| L
1894 |
conjugate |
|
|
Gold, 10nm |
| L
1882 |
conjugate |
|
|
Agarose |
| Ulex
Europaeus |
Top
of Page |
| L
5505 |
UEA I |
68 |
|
H |
a-L-fuc |
|
|
| L
8146 |
conjugate |
|
|
Peroxidase |
| L
8262 |
conjugate |
|
|
Biotin |
| L
9006 |
conjugate |
|
|
FITC |
| L
4889 |
conjugate |
|
|
TRITC |
| L
6263 |
Vicia faba |
50 |
4(a&ß)a |
|
man, glc |
(+) |
|
| L
4011 |
Vicia villosa |
139 |
4a |
A1+Tn
|
galNAc |
|
|
| L
9388 |
conjugate |
|
|
Agarose |
| L
7513 |
Isolectin B4 |
143 |
4 |
Tn |
galNAc |
|
|
| L
7888 |
conjugate |
|
|
Agarose |
| L
2662 |
Viscum album |
115g |
4(a&ß)a |
|
ß-gal |
|
|
| |
Wisteria floribunda |
68 |
2 |
|
galNAc |
|
|
| L
1516 |
conjugate |
|
|
Biotin |
| L
2016 |
Reduced |
34 |
1 |
|
galNAc |
|
|
| L
1766 |
conjugate |
|
|
Biotin |
| NOTES: |
|
a. Subunits are of different molecular
weights
b. Mitogenic for neuraminidase-treated lymphocytes
c. Concentration-dependent mol. wt. change
d. Non-agglutinating and mitogenic |
e. Inhibits mitogenic activity of PHA
f. Data given for PWM Pa2
g. Data given for VAA(I)
h. Agglutinates rabbit, but not human, erythrocytes |