Order Center
Biochemicals
Antibiotic Explorer
BioUltra
Biological Detergents
Detection
Enzyme Explorer
Lipids
Microparticles
Calibration Size Standards
Coupling to Microparticles
Dyed Microparticles
Fluorescent Microparticles
Magnetic Microparticles
Microparticles Synthesis
Microparticles in Microscopy
Monodisperse Microparticles
Silica Microparticles
Washing Microparticles
Peptide Explorer
Pharmacopoeia
PolyAmino Acids
 Dyed Microparticles
Microparticles
A number of dyed microparticles based on polystrene are also available. Dyes include dark blue and dark red. Other properties are the same as for the non-dyed particles:
  • Density: 1.05 g/cm3
  • Refractive Index: 1.59
  • High monodispersity and uniform spherical shape
  • Hydrophobic surface
  • Non-specific adsorption of proteins
  • Low temperature resistance up to 100°C
  • Soluble in organic solvents (dependent on the degree of cross-linking)
  • Swellable in organic solvents
  • Coefficient of Variation (C.V. value) of <3 % for research grade particles and for particle size standards available
Dyed Microparticles Based on Polystyrene