|
In addition to the polymer based particles we now offer a range of silica particles offering unique properties:
- Density: 1.8-2 g/cm3
- Refractive Index: 1.37
- Hydrophilic anionic surface
- Narrow size distribution
- Temperature resistance up to 1000°C
- Good mechanical stability
- Stable in organic solvents
- Soluble in strong bases and in hydrofluoric acid
Microparticles based on silica
|