Order Center
Product Catalog →  Cell Biology →  Cell Signaling and Neuroscience →  Peptides and Proteins →  Peptides →  Neuropeptides →  Bombesins
 Print Preview
Bombesins
Bombesin is a tetradecapeptide originally isolated from the skin of the frog Bombina bombina. Bombesin acts at specific brain nuclei to inhibit food intake. It also increases blood glucose levels.
Description
Biochem/physiol Actions
Product #
Bombesin acetate hydrate ≥97% (HPLC) Tetradecapeptide originally isolated from the skin of the frog Bombina bombina. Bombesin acts at specific brain nuclei to inhibit food intake. It also increases blood glucose levels.
B4272
Bombesin acetate hydrate BioChemika, ≥97.0% (HPLC) 15560
[Lys3]-Bombesin ≥97% (HPLC) B1647
[Tyr4]-Bombesin ≥97% (HPLC) B5397
Gastrin Releasing Peptide human ≥97% (HPLC) G8022
β-(2-Naphthyl)-D-Ala-Cys-Tyr-D-Trp-Orn-Val-Cys-β-(2-Naphthyl)-D-Ala-NH2 [disulfide bridge: 2-7] >97% (HPLC), solid Selective neuromedin B receptor antagonist.
N3280
Neuromedin B ≥90% (HPLC) Bombesin-like peptide that is present in the mammalian central nervous system and gastrointestinal tract that has been implicated in the control of food intake, smooth muscle contraction, and thermoregulation. It is also an autocrine growth factor in non-small cell lung cancer.
N3762
[Ser2]-Neuromedin C ≥97% (HPLC) N1773
Neuromedin U-25 human ≥97% (HPLC), lyophilized powder N4284
Neuromedin U-25 porcine ≥97% (HPLC), solid Potent stimulator of rat uterus smooth muscle.
N8138
Neuromedin U rat ≥97% (HPLC), lyophilized powder N4409
RC-3095 ≥98% (HPLC), powder RC-3095 is a potent BB2 (GRP-preferring) bombesin receptor antagonist.
R9653
 Site Use Terms Terms and Conditions of Sale Privacy
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.