| Other Enzyme Inhibitors |
|
| Product Name |
Product # |
Specificity |
| b-Chloro-L-alanine hydrochloride |
C9033 |
Alanine aminotransferase |
| L-Cycloserine |
C1159 |
Alanine hydroxymethyltransferase, D- |
| DL-Cycloserine Crystalline |
C7005 |
Alanine hydroxymethyltransferase, D- |
| Phebestin >99% (HPLC), Lyophilized powder (From acetic acid solution.) |
P7240 |
Aminopeptidase N |
| Apstatin approx. 95% (HPLC) |
A1395 |
Aminopeptidase P |
| Angiotensin Converting Enzyme Inhibitor minimum 95% (TLC) |
A0773 |
Angiotensin Converting Enzyme |
| Bradykinin potentiator B minimum 97% (TLC) |
B0507 |
Angiotensin Converting Enzyme |
| Bradykinin potentiator C minimum 97% (TLC) |
B0632 |
Angiotensin Converting Enzyme |
| N-Succinyl-L-proline |
S6633 |
Angiotensin Converting Enzyme |
| Ne-Trifluoroacetyl-Lys-Pro |
T7025 |
Angiotensin Converting Enzyme |
| E-64 |
E3132 |
Bromelain |
| Na-p-Tosyl-L-lysine chloromethyl ketone hydrochloride minimum 97% (TLC), Powder |
T7254 |
Bromelain |
| Calcineurin autoinhibitory fragment minimum 90% (HPLC) |
C3937 |
Calcineurin (phosphatase activity) |
| Autocamtide 2-related inhibitory peptide trifluoroacetate salt minimum 97% (HPLC), Lyophilized powder |
A4308 |
Calmodulin dependent Protein Kinase II |
| Calmodulin Kinase II Inhibitor from rat minimum 70%, recombinant, expressed in Escherichia coli (histidine-tagged) |
C1360 |
Calmodulin dependent Protein Kinase II |
| KN-62 approx. 95%, Solid |
I2142 |
Calmodulin dependent Protein Kinase II |
| Antipain hydrochloride microbial |
A6191 |
Calpain |
| Acetyl-Calpastatin (184-210) Human approx. 95% (HPLC), Powder |
C4285 |
Calpain |
| Z-Leu-Leu-Tyr-fluoromethyl ketone minimum 90% (TLC), Lyophilized powder |
C9234 |
Calpain |
| Z-Leu-Leu-Tyr-CHN2 minimum 98% (TLC), Solid |
C9859 |
Calpain |
| E-64c |
E0514 |
Calpain |
| E-64d |
E8640 |
Calpain |
| Leupeptin trifluoroacetate salt minimum 90% (HPLC), microbial |
L2023 |
Calpain |
| N-Methoxysuccinyl-Val-HOMO-Phe-fluoromethyl ketone minimum 95% (TLC), Powder |
M5439 |
Calpain |
| Calpain Inhibitor II minimum 97% (TLC), Powder |
A6060 |
Calpain I |
| Calpain Inhibitor I minimum 97% (TLC), Powder |
A6185 |
Calpain I |
| Calpain Inhibitor Peptide minimum 95% (HPLC) |
C9181 |
Calpain I |
| Calpain Inhibitor II minimum 97% (TLC), Powder |
A6060 |
Calpain II |
| Calpain Inhibitor I minimum 97% (TLC), Powder |
A6185 |
Calpain II |
| Antipain hydrochloride microbial |
A6191 |
Calpain II |
| Acetyl-Calpastatin (184-210) Human approx. 95% (HPLC), Powder |
C4285 |
Calpain II |
| Calpain Inhibitor Peptide minimum 95% (HPLC) |
C9181 |
Calpain II |
| E-64c |
E0514 |
Calpain II |
| Leupeptin trifluoroacetate salt minimum 90% (HPLC), microbial |
L2023 |
Calpain II |
| Z-L-Phe chloromethyl ketone |
C9511 |
Chymotrypsin A gamma, Bovine |
| N-p-Tosyl-L-phenylalanine chloromethyl ketone minimum 97% (TLC), Powder |
T4376 |
Chymotrypsin, alpha- |
| Histatin 5 minimum 97% (HPLC) |
H6027 |
Clostripain |
| Na-p-Tosyl-L-lysine chloromethyl ketone hydrochloride minimum 97% (TLC), Powder |
T7254 |
Clostripain |
| 5-(R,S)-T-trans-cinnamido-7-methyl-4-oxo-octanoyl-Pro-Pro minimum 90% (NMR, TLC) |
C0360 |
Collagenase |
| Isoamylphosphonyl-Gly-Pro-Ala dipotassium salt 95% (HPLC, NMR), Lyophilized powder |
I7029 |
Collagenase |
| N-Phospho-Ile(O-ethyl)-Tyr(O-benzyl)-Gly dipotassium salt 95% (TLC) |
P6115 |
Fibroblast collagenase |
| Ile-Pro-Ile minimum 97% (HPLC) |
I9759 |
Dipeptidyl peptidase IV |
| Eglin C fragment 60-63 methyl ester minimum 97% (HPLC) |
E2017 |
Elastase, leukocyte |
| N-Acetyl-eglin C recombinant, expressed in unspecified host |
E7888 |
Elastase, leukocyte |
| Cys-Val-2-Nal-Met minimum 90% (HPLC) |
C4433 |
farnesyltransferase, p21ras |
| Cys-4-(2-aminobenzoyl)-Met minimum 90% (HPLC) |
C5684 |
Farnesyltransferase, Ras |
| E-64 |
E3132 |
Ficin |
| Z-Ala-Ala-Asp(OMe)-chloromethyl ketone minimum 98% (TLC), Solid |
C0235 |
Granzyme B |
| His-Cys-Lys-Phe-Trp-Trp minimum 95% (HPLC) |
H6387 |
HIV integrase |
| Bestatin hydrochloride minimum 98% (HPLC) |
B8385 |
Leucine Aminopeptidase |
| Amastatin hydrochloride minimum 97% (HPLC) |
A1276 |
Leucine aminopeptidase, cytosol |
| NG-Hydroxy-L-arginine acetate salt |
H7278 |
Nitric Oxide Synthase |
| L-N6-(1-Iminoethyl)lysine hydrochloride |
I8021 |
Nitric Oxide Synthase |
| L-N5-(1-Iminoethyl)ornithine hydrochloride |
I8768 |
Nitric Oxide Synthase |
| Nw-Nitro-L-arginine minimum 98% (TLC) |
N5501 |
Nitric Oxide Synthase |
| Nw-Nitro-L-arginine methyl ester hydrochloride minimum 98% (TLC), Powder |
N5751 |
Nitric Oxide Synthase |
| NG-Methyl-L-arginine acetate salt approx. 98% (TLC) |
M7033 |
Nitric Oxide Synthase (iNOS) |
| E-64 |
E3132 |
Papain |
| Gly-Gly-Tyr-Arg minimum 97% (HPLC) |
G5386 |
Papain |
| Leupeptin trifluoroacetate salt minimum 90% (HPLC), microbial |
L2023 |
Papain |
| Na-p-Tosyl-L-lysine chloromethyl ketone hydrochloride minimum 97% (TLC), Powder |
T7254 |
Papain |
| N-Acetyl-Glu-Ser-Met-Asp-al minimum 90% (HPLC), Powder |
A1341 |
Pro-caspase 3 |
| N-Acetyl-Ile-Glu-Thr-Asp-al approx. 99% (TLC), Powder |
A1216 |
Pro-caspase 3 |
| 2-Aminobenzoyl-Ala-Phe-Ala-Phe-Asp-Val-Phe-nitro-Tyr-Asp trifluoroacetate salt >90% (HPLC) |
A9971 |
Asp-specific proteases |
| a-Boc-Deacetylleupeptin minimum 97% (TLC) |
B7530 |
Protease (Non-selective serine and cysteine protease inhibitor) |
| Acetyl-pepstatin minimum 95% (HPLC) |
A4815 |
Protease HIV-1ANT |
| Histatin 5 minimum 97% (HPLC) |
H6027 |
Protease of Bacteroides gingivalis |
| Propionyl-leupeptin hemisulfate minimum 90% (HPLC) |
L3402 |
Proteases, serine and cysteine |
| Z-Leu-Leu-Phe-CHO >95% (HPLC), Solid |
C0358 |
Proteasome inhibitor |
| Z-Leu-Leu-Leu-B(OH)2 >95% (HPLC), Solid |
C9857 |
Proteasome inhibitor |
| Pepstatin A microbial, minimum 90% (HPLC) |
P5318 |
Renin |
| D-His-Pro-Phe-His-Leu-y-(CH2NH)-Leu-Val-Tyr minimum 90% (HPLC) |
H6137 |
Renin, human |
| [Asn670, Sta671, Val672]-Amyloid b/A4 Precursor Protein 770 Fragment 662-675 trifluoroacetate salt minimum 95% (HPLC) |
A1847 |
APP-Secretase |
| Glu-Val-Asn-[(2R,4S,5S)-5-amino-4-hydroxy-2,7-dimethyloctanoyl]-Ala-Glu-Phe trifluoroacetate salt >93% (HPLC), Solid |
G8291 |
beta-Secretase |
| [(2R,4R,5S)-2-Benzyl-5-(Boc-amino)-4-hydroxy-6-phenylhexanoyl]-Leu-Phe-NH2 trifluoroacetate salt >90% (TLC) |
B5306 |
gammaSecretase |
| N-Acetyl-eglin C recombinant, expressed in unspecified host |
E7888 |
Thrombin |
| Leupeptin trifluoroacetate salt minimum 90% (HPLC), microbial |
L2023 |
Thrombin |
| Na-p-Tosyl-L-lysine chloromethyl ketone hydrochloride minimum 97% (TLC), Powder |
T7254 |
Thrombin |
| D-Val-Leu-Lys-chloromethyl ketone minimum 90% (HPLC), White powder |
V3763 |
Thrombin |
| Glu-Gly-Arg chloromethyl ketone minimum 90% (HPLC), Powder |
G2917 |
Urokinase |
| Other Enzyme Substrates |
| Kinase and Phosphatase Substrates |
|
back to top |
| Product Name |
Product # |
Specificity |
| NFAT 1 from mouse N-terminal 6-His tagged, Truncated sequence amino acids 1-297, minimum 90% (SDS-PAGE), Solution, recombinan |
N9659 |
SAPK2a/p38a |
| Protein Tyrosine Phosphatase Substrate Monophosphate minimum 97% (HPLC) |
P6462 |
Protein Tyrosine Phosphatase |
| L-a-Phosphatidyl-D-myo-inositol 3,4,5-triphosphate, dioctanoyl |
P9953 |
PTEN |
| LAT human recombinant, expressed in Escherichia coli, approx. 50%, Buffered aqueous glycerol solution |
L9413 |
ZAP 70 Kinase |
| Protease Substrates |
|
back to top |
| Product Name |
Product # |
Enzyme |
| N-Benzoyl-Pro-Phe-Arg-p-nitroanilide hydrochloride |
B2133 |
Agkistrodon |
| Z-Val-Lys-Met 7-amido-4-methylcoumarin |
C2681 |
Amyloid A4-generating enzymes |
| 4-Nitrophenyl a-L-arabinofuranoside |
N3641 |
Arabinofuranosidase III, alpha-L- |
| 4-Methylumbelliferyl a-L-arabinopyranoside |
M3167 |
Arabinosidase |
| Z-L-Lys-ONp hydrochloride |
C3637 |
Bromelain |
| Autocamtide 2 trifluoroacetate salt minimum 97% (HPLC), Powder |
A0591 |
Calmodulin-dependent protein kinase II |
| Syntide 2 minimum 97% (HPLC) |
S2525 |
Calmodulin-dependent protein kinase II |
| Calcium/calmodulin-dependent protein kinase II-d fragment 345-358 |
C0843 |
Calmodulin-dependent protein kinase IV |
| N-Succinyl-Leu-Leu-Val-Tyr-7-Amido-4-Methylcoumarin minimum 90% (HPLC) |
S6510 |
Calpain |
| N-Succinyl-Leu-Tyr-7-amido-4-methylcoumarin |
S1153 |
Calpain |
| Hippuryl-Lys |
H6750 |
Carboxypeptidase B |
| N-(3-[2-Furyl]acryloyl)-Ala-Lys Powder |
F5882 |
Carboxypeptidase N |
| Acetyl-Lys-D-Ala-D-Ala minimum 95% (HPLC) |
A6950 |
Carboxypeptidase G |
| Na,Ne-Diacetyl-Lys-D-Ala-D-lactic acid |
D2279 |
Carboxypeptidase, penicillin-sensitive D-alanine |
| Na,Ne-Diacetyl-Lys-D-Ala-D-Ala |
D9904 |
Carboxypeptidase, penicillin-sensitive D-alanine |
| N-(3-[2-Furyl]acryloyl)-Phe-Phe |
F7133 |
Carboxypeptidase, Serum |
| Hippuryl-L-phenylalanine |
H6875 |
CarboxypeptidaseA |
| Ala-Ala-Phe-7-amido-4-methylcoumarin |
A3401 |
Chymotrypsin, alpha- |
| Ala-Ala-Val-Ala p-nitroanilide |
A9273 |
Chymotrypsin, alpha- |
| N-Benzoyl-L-tyrosine ethyl ester |
B6125 |
Chymotrypsin, alpha- |
| N-Benzoyl-L-tyrosine p-nitroanilide |
B6760 |
Chymotrypsin, alpha- |
| Glutaryl-Ala-Ala-Phe-4-methoxy-b-naphthylamide |
G3769 |
Chymotrypsin, alpha- |
| N-Glutaryl-Gly-Gly-Phe-b-naphthylamide |
G4636 |
Chymotrypsin, alpha- |
| 4-Methylumbelliferyl p-trimethylammoniocinnamate chloride Powder |
M4507 |
Chymotrypsin, alpha- |
| N-Succinyl-Gly-Gly-Phe-p-nitroanilide |
S1899 |
Chymotrypsin, alpha- |
| N-Succinyl-L-phenylalanine-p-nitroanilide |
S2628 |
Chymotrypsin, alpha- |
| N-Succinyl-Ala-Ala-Pro-Phe 4-methoxy-2-naphthylamide minimum 98% (TLC), Solid |
S5314 |
Chymotrypsin, alpha- |
| N-Succinyl-Ala-Ala-Pro-Phe p-nitroanilide |
S7388 |
Chymotrypsin, alpha |
| N-Succinyl-Ala-Ala-Pro-Phe-7-amido-4-methylcoumarin |
S9761 |
Chymotrypsin, alpha |
| N-Succinyl-Ala-Ala-Pro-Phe 7-amido-4-trifluoromethylcoumarin minimum 99% (TLC), Solid |
S5189 |
Chymotrypsin, alpha- |
| Z-Gly-Pro-Leu-Gly-Pro |
C0627 |
Collagenase |
| Collagen Type II-FITC Conjugate bovine articular cartilage approx. 1 mg/ml, Solution |
C4486 |
Collagenase |
| Collagen Type I-FITC Conjugate bovine skin approx. 1 mg/ml, Solution |
C4361 |
Collagenase |
| Collagen-Fluorescein bovine |
C5858 |
Collagenase |
| Z-Gly-Pro-Gly-Gly-Pro-Ala |
C8008 |
Collagenase |
| N-(2,4-Dinitrophenyl)-Pro-Leu-Gly-Cys(Me)-His-Ala-D-Arg amide minimum 95% (HPLC) |
D0296 |
Collagenase |
| N-[3-(2-Furyl)acryloyl]-Leu-Gly-Pro-Ala |
F5135 |
Collagenase |
| 7-Methoxycoumarin-4-acetyl-Pro-(cyclohexyl)-Ala-Gly-Nva-His-Ala-L-(2,4-di nitrophenyl)diaminoproponic acid Solid |
M3314 |
Collagenase-3 |
| Boc-Leu-Gly-Arg-7-amido-4-methylcoumarin acetate salt |
B4511 |
Convertase |
| N-Acetyl-Gly(t-butyl)-Gly(t-butyl)-Asn(Methyl)2-Ala-7-amido-4- methylcoumarin minimum 97% (HPLC), Solid |
A2347 |
Cytomegalovirus (HCMV) protease |
| 2-Aminobenzoyl-Gly(t-butyl)-Asn(methyl)2-Ala-Ser-Ser-Arg-Leu-T yr(NO2)-Tyr-Arg trifluoroacetate salt >94% (HPLC) |
A9846 |
Cytomegalovirus (HCMV) protease |
| Lys-Ala-7-amido-4-methylcoumarin dihydrochloride |
L8139 |
Dipeptidyl aminopeptidase |
| Gly-Arg 4-methoxy-b-naphthylamide dihydrochloride |
G1013 |
Dipeptidyl aminopeptidase I |
| Lys-Ala 4-methoxy-b-naphthylamide dihydrochloride |
L2270 |
Dipeptidyl aminopeptidase II |
| Arg-Arg b-naphthylamide trihydrochloride |
A3261 |
Dipeptidyl aminopeptidase III |
| Arg-Arg 4-methoxy-b-naphthylamide trihydrochloride |
A9398 |
Dipeptidyl aminopeptidase III |
| Arg-Pro p-nitroanilide acetate salt |
A1204 |
Dipeptidyl aminopeptidase IV |
| Gly-Pro 4-methoxy-b-naphthylamide |
G9137 |
Dipeptidyl aminopeptidase IV |
| Gly-Pro 4-methoxy-b-naphthylamide hydrochloride Powder |
G9262 |
Dipeptidyl aminopeptidase IV |
| Gly-Pro p-nitroanilide hydrochloride |
G0513 |
Dipeptidyl peptidase IV |
| Gly-Pro-7-amido-4-methylcoumarin hydrobromide |
G2761 |
Dipeptidyl peptidase IV. |
| Gly-Pro p-nitroanilide p-toluenesulfonate salt |
G2901 |
Dipeptidyl peptidase IV. |
| N-Succinyl-Ala-Ala-Ala-p-nitroanilide |
S4760 |
Elastase |
| Boc-D-Phe-Ala-Nle-p-nitroanilide |
B8397 |
Elastase, Hunan Granulocyte |
| Boc-Ala-Pro-Nva p-chlorothiobenzyl ester |
B2017 |
Elastase, leukocyte |
| N-Methoxysuccinyl-Ala-Ala-Pro-Val-7-amido-4-trifluoromethylcoumarin Solid |
M4563 |
Elastase, leukocyte |
| N-(Methoxysuccinyl)-Ala-Ala-Pro-Val 4-nitroanilide |
M4765 |
Elastase, leukocyte |
| N-Methoxysuccinyl-Ala-Ala-Pro-Val-7-amido-4-methylcoumarin |
M9771 |
Elastase, leukocyte |
| N-Methoxysuccinyl-Ala-Ala-Pro-Val-7-amido-4-methylcoumarin |
M9771 |
Elastase, pancreatic |
| 7-Methoxycoumarin-4-acetyl [Ala7-(2,4-Dinitrophenyl)Lys9]-Bradykinin trifluoroacetate salt >94% (HPLC) |
B5181 |
Endothelin-converting enzyme-1 |
| N-Dansyl-D-Ala-Gly-p-nitro-Phe-Gly approx. 95% (HPLC) |
D2155 |
Enkephalinase |
| Glutaryl-Ala-Ala-Phe-4-methoxy-b-naphthylamide |
G3769 |
Enkephalinase |
| Gly-Asp-Asp-Asp-Asp-Lys-b-naphthylamide minimum 95% (HPLC) |
G5261 |
Enterokinase |
| N-Benzoyl-Ile-Glu-Gly-Arg p-nitroanilide hydrochloride |
B2291 |
Factor X Activated (Xa) |
| N-Benzoyl-Ile-Glu-Gly-Arg p-nitroanilide acetate salt |
B7020 |
Factor X Activated (Xa) |
| Boc-Ile-Glu-Gly-Arg-7-amido-4-methylcoumarin hydrochloride |
B9760 |
Factor X Activated (Xa) |
| Factor Xa chromogenic substrate Solid |
F3301 |
Factor X Activated (Xa) |
| 4-Methylumbelliferyl 4-guanidinobenzoate hydrochloride monohydrate Crystalline |
M4149 |
Factor X Activated (Xa) |
| Boc-g-benzyl-Glu-Ala-Arg-7-amido-4-methylcoumarin hydrochloride approx. 95% |
B4903 |
Factor Xia |
| Boc-Phe-Ser-Arg-7-amido-4-methylcoumarin |
B6388 |
Factor XIA |
| N-Acetyl-Arg-Glu-Lys-Arg-7-amido-4-trifluoromethylcoumarin >97% (HPLC), Solid |
C6483 |
Furin |
| [2-Aminobenzoyl-Gly1]-Galanin fragment (1-10)-Lys(retro-m-Nitro-Tyr) amide, human trifluoroacetate salt >95% (HPLC), Solid |
G8916 |
Galanin degrading zinc-metallopeptidase |
| Ras, modified human recombinant, expressed in Escherichia coli, approx. 95% (SDS-PAGE), Buffered aqueous glycerol solution |
R0145 |
Geranylgeranyltransferase |
| Rac-1 protein human recombinant, expressed in Escherichia coli, minimum 85%, Buffered aqueous glycerol solution |
R3012 |
Geranylgeranyltransferase |
| Boc-Val-Leu-Gly-Arg p-nitroanilide hydrobromide |
B8391 |
Horseshoe crab clotting enzyme |
| Interleukin-1b precursor (pIL-1b) fragment 110-123 minimum 97% (HPLC) |
I2775 |
Interleukin-1beta converting enzyme |
| N-Benzoyl-Pro-Phe-Arg-p-nitroanilide hydrochloride |
B2133 |
Kallikrein |
| Na-Benzoyl-L-arginine methyl ester bicarbonate salt |
B4256 |
Kallikrein |
| Z-Phe-Arg 7-amido-4-methylcoumarin hydrochloride |
C9521 |
Kallikrein |
| Kininogen, low molecular weight from human plasma >95% (SDS-PAGE), Lyophilized powder (containing sodium acetate and sodium chloride) |
K3628 |
Kallikrein |
| D-Val-Leu-Arg 7-amido-4-methylcoumarin Solid |
V2763 |
Kallikrein |
| D-Val-Leu-Arg 7-amido-4-trifluoromethylcoumarin minimum 90% (HPLC), Solid |
V2888 |
Kallikrein |
| L-Leucine p-nitroanilide |
L9125 |
Leucine aminopeptidase |
| N-Succinyl-Ala-Ala-Pro-Phe-7-amido-4-methylcoumarin |
S9761 |
Leucine Aminopeptidase |
| N-Succinyl-Phe-Gly-Leu-b-naphthylamide |
S6768 |
LH-RH-degrading endopeptidase |
| DABCYL-[Nle1077]-Collagen Type III a 1 Chain fragment 1070-1077EDANS, Human trifluoroacetate salt >96% (HPLC), Type III a 1 chain |
C9732 |
Metalloproteinase |
| 7-Methoxycoumarin-4-acetyl-Arg-Pro-Leu-Ala-Leu-Trp-Arg-L-a,b-diaminopropionyl(2,4-dinitrophenyl) amide >97% (HPLC), Solid |
M0813 |
Metalloproteinase |
| N-(2,4-Dinitrophenyl)-Pro-Leu-Gly-Met-Trp-Ser-Arg Solid |
D4315 |
Matrix metalloproteinase |
| 7-Methoxycoumarin-4-acetyl-Pro-Leu-Gly-Leu-b-(2,4-dinitrophenylamino)Ala-Ala-Arg amide minimum 97% (HPLC) |
M6412 |
Matrix metalloproteinase |
| 7-Methoxycoumarin-4-acetyl-Pro-Leu-Ala-Nva-L-(2,4-dinitrophenyl)diaminopr opionyl-Ala-Arg-amide Solid |
M3439 |
Matrix metalloproteinase-3 |
| N-(2,4-Dinitrophenyl)-Pro-Leu-Gly-Leu-Trp-Ala-D-Arg amide |
D2293 |
Matrix metalloproteinases MMP-1 and MMP-9 |
| N-(2,4-Dinitrophenyl)-Pro-Leu-Gly-Leu-Trp-Ala-D-Arg amide |
D2293 |
Matrix metalloproteinases MMP-1 and MMP-9 |
| Na-Benzoyl-L-arginine-7-amido-4-methylcoumarin hydrochloride |
B7260 |
Papain |
| Fibrin Blue substrate for pepsin at low pH (High blanks result at high pH) |
F5255 |
Pepsin |
| N-Succinyl-Ala-Ala-Pro-Phe p-nitroanilide |
S7388 |
Peptidyl prolyl isomerase |
| D-Ala-Leu-Lys-7-amido-4-methylcoumarin 95% (HPLC) |
A8171 |
Plasmin |
| Boc-Val-Leu-Lys-7-amido-4-methylcoumarin |
B1136 |
Plasmin |
| Z-Ala-Ala-Lys 4-methoxy-b-naphthylamide formate salt |
C2147 |
Plasmin |
| Gly-Arg-p-nitroanilide dihydrochloride |
G8148 |
Plasmin |
| D-Ile-Phe-Lys p-nitroanilide minimum 95% (HPLC) |
I6886 |
Plasmin |
| N-p-Tosyl-Gly-Pro-Lys 4-nitroanilide acetate salt |
T6140 |
Plasmin |
| D-Val-Leu-Lys 4-nitroanilide dihydrochloride |
V0882 |
Plasmin |
| N-Methoxysuccinyl-Phe-Leu-Phe-7-amido-4-trifluoromethylcoumarin Solid |
M4813 |
ATP-dependent protease |
| N-Methoxysuccinyl-Phe-Leu-Phe 4-methoxy-2-naphthylamide minimum 90%, Solid |
M4938 |
ATP-dependent protease |
| 2-Aminobenzoyl-Gly-Ala-Ala-Pro-Phe-nitro-Tyr-Asp trifluoroacetate salt minimum 90% (HPLC) |
A0222 |
Proline SpecificEndopeptidase |
| Anthranilyl-Asp-Phe-Arg-Leu-Phe-Ala-Phe-nitro-Tyr-Asp minimum 97% (HPLC) |
A0324 |
Protease |
| Azoalbumin |
A2382 |
Protease |
| Azocasein |
A2765 |
Protease |
| Albumin, Fluorescein isothiocyanate Conjugate bovine |
A9771 |
Protease |
Azo dye-impregnated collagen non-specific protease substrate rate of dye release may vary from lot to lot (Dye-impregnated hide powder.) |
A4341 |
Protease |
| Z-Gly-Gly-Leu p-nitroanilide |
C3022 |
Protease |
| N-(3-[2-Furyl]acryloyl)-Ala-Phe amide |
F2380 |
Protease |
| Fibrin Blue substrate for pepsin at low pH (High blanks result at high pH) |
F5255 |
Protease |
| N-(3-[2-Furyl]acryloyl)-Gly-Leu amide |
F7383 |
Protease |
| L-Leucine-7-amido-4-methylcoumarin hydrochloride |
L2145 |
Protease |
| N-Succinyl-Gly-Gly-Phe-p-nitroanilide |
S1899 |
Protease B |
| Anthranilyl-Ala-Phe-Ala-Phe-Glu-Val-Phe-nitro-Tyr-Asp minimum 90% (HPLC) |
A3567 |
Proteases glutamate-specific |
| Z-Leu-Leu-Glu-7-amido-4-methylcoumarin minimum 95%, Solid |
C0483 |
Proteasome |
| Z-Leu-Leu-Leu-7-amido-4-methylcoumarin minimum 95% (TLC and HPLC), Solid |
C0608 |
Proteasome |
| N-Succinyl-Leu-Leu-Val-Tyr-7-amido-4-trifluoromethylcoumarin minimum 98% (TLC), Solid |
S4939 |
Proteasome |
| Boc-Leu-Ser-Thr-Arg-7-amido-4-methylcoumarin |
B4636 |
Protein C. |
| Renin Substrate 1 |
R8276 |
Renin |
| Arg-Glu(EDANS)-[Asn670, Leu671]-Amyloid b/A4 Precursor Protein 770 Fragment 669-674-(DABCYL)Lys-Arg minimum 96% (HPLC), Solid |
A2847 |
pro-beta-secretase (pro-memapsin 2) |
| Mca-[Asn670, Leu671]-A4 Precursor Protein 770 fragment 667-675-(Dnp)Lys amide >96% (HPLC), Solid |
M1063 |
pro-beta-secretase (pro-memapsin 2) |
| 7-Methoxycoumarin-4-acetyl-[Asn670, Leu671]-Amyloid b/A4 Precursor Protein 770 Fragment 667-674-(2,4-dinitrophenol)Dap minimum 97% (HPLC) |
A1222 |
beta-Secretase |
| 7-Methoxycoumarin-4-acetyl[Asn670,Leu671]-Amyloid b/A4 Precursor Protein 770 (667-676)-(2,4-dinitrophenyl)Lys-Arg-Arg Amide minimum 96% (HPLC) |
A1347 |
beta-Secretase |
| 7-Methoxycoumarin-4-acetyl-[Asn670, Leu671]-Amyloid b/A4 Precursor Protein 770 Fragment 667-676-(2,4-dinitrophenyl)Lys-Arg-Arg amide minimum 90% (HPLC) |
A1472 |
beta-Secretase |
| 7-Methoxycoumarin-4-acetyl-[Asn670, Leu671]-Amyloid b/A4 Precursor Protein 770 Fragment 669-674-(2,4-dinitrophenyl)Lys trifluoroacetate salt minimum 96% (HPLC) |
A1597 |
beta-Secretase |
| [Asn670, Leu671]-Amyloid b/A4 Precursor Protein 770 Fragment 667-676 trifluoroacetate salt minimum 95% (HPLC), Solid |
A1972 |
beta-Secretase |
| [Val671]-Amyloid b/A4 Precursor Protein 770 Fragment 667-676 trifluoroacetate salt minimum 95% (HPLC), Solid |
A2722 |
beta-Secretase |
| Amyloid b/A4 Precursor Protein 770 Fragment 667-676 minimum 96% (HPLC), Solid |
A3222 |
beta-Secretase |
| DABCYL-[Asn670, Leu671]-Amyloid b/A4 Precursor Protein 770 fragment 661-675-EDANS trifluoroacetate salt minimum 97% (HPLC), Solid |
A4972 |
beta-Secretase |
| DABCYL-[Asn670, Leu671]-Amyloid b/A4 Precursor Protein 770 Fragment 669-674-EDANS trifluoroacetate salt minimum 95% (HPLC), Solid |
A5097 |
beta-Secretase |
| 7-Methoxycoumarin-4-acetyl[Asn670,Leu671]-Amyloid b/A4 Precursor Protein 770 (667-676)-(2,4-dinitrophenyl)Lys-Arg-Arg Amide minimum 96% (HPLC) |
A1347 |
Thimet oligopeptidase (TOP) |
| 7-Methoxycoumarin-4-acetyl-[Asn670, Leu671]-Amyloid b/A4 Precursor Protein 770 Fragment 667-676-(2,4-dinitrophenyl)Lys-Arg-Arg amide minimum 90% (HPLC) |
A1472 |
Thimet oligopeptidase (TOP) |
| N-Benzoyl-Phe-Val-Arg 4-methoxy-b-naphthylamide hydrochloride |
B1260 |
Thrombin |
| Boc-b-benzyl-Asp-Pro-Arg-7-amido-4-methylcoumarin hydrochloride approx. 95% |
B4028 |
Thrombin |
| N-Benzoyl-Phe-Val-Arg-p-nitroanilide hydrochloride hydrate |
B7632 |
Thrombin |
| Boc-Val-Pro-Arg-7-amido-4-methylcoumarin hydrochloride |
B9385 |
Thrombin |
| 4-Methylumbelliferyl 4-guanidinobenzoate hydrochloride monohydrate Crystalline |
M4149 |
Thrombin |
| Sar-Pro-Arg p-nitroanilide dihydrochloride |
S9009 |
Thrombin |
| N-p-Tosyl-Gly-Pro-Arg 7-amido-4-methylcoumarin hydrochloride |
T0273 |
Thrombin |
| N-p-Tosyl-Gly-Pro-Arg p-nitroanilide acetate salt |
T1637 |
Thrombin |
| Thrombin generation chromogenic substrate minimum 90% (TLC), Solid |
T3068 |
Thrombin |
| pGlu-Gly-Arg-7-amido-4-trifluoromethylcoumarin minimum 95% (HPLC), Lyophilized powder |
P5615 |
Tissue Plasinogen Activator |
| D-Val-Phe-Lys p-Nitroanilide dihydrochloride minimum 97% (HPLC) |
V6884 |
Tissue Plasinogen Activator |