Sigma-Aldrich
Biochemicals
Antibiotic Explorer
BioUltra
Biological Detergents
Detection
Enzyme Explorer
Lipids
Microparticles
Peptide Explorer
Pharmacopoeia
Product Index
PolyAmino Acids
SyntheChol™ From Sigma-Aldrich
The First Non-animal Derived Synthetic Cholesterol
US Customers Only
Animal source cholesterol could contain allergens, viruses and TSEs (transmissible spongiform encephalopathies) rendering it problematic for use in drug delivery and pharmaceutical applications. Sigma-Aldrich was the first to see the need and manufacture Synthechol™, synthetic non-animal source cholesterol. Ideal for liposomes used in drug delivery, gene therapy and other pharmaceutical applications.
Features
  • Synthesized from plant derived raw materials
  • High Quality
  • Sigma-Aldrich is the sole supplier for synthetic cholesterol
  • Commercial quantities available
  • Can be custom manufactured (e.g. derivatized) or packaged per your specifications
  • Reduced production costs allow us to pass savings to you
Order Information
Product # Product DescriptionPackage SizeOld Pricing ($)New Lower Pricing ($)
C1231-10g SyntheChol™ (cGMP manufactured) 10 g 750.00 700.00
C1231-100 g SyntheChol™ (cGMP manufactured) 100 g 5000.00 4500.00
C9913-50 mg Cholesterol non-animal source –synthetic (research use only) 50 mg 72.10 25.00
C9913-500 mg Cholesterol non-animal source –synthetic (research use only) 500 mg 396.35 100.00
For inquiries on commercial quantities, please call SAFC at 800-244-1173, e-mail: safcglobal@sial.com, or contact your local SAFC sales representative.