Amino Acids Reference Chart |
 |
PolyAmino Acids |
 |
 |
| |
|
|
Neutral - polar side chains |
| |
 |
| Asparagine, Asn, N |
|
 |
| Cysteine, Cys, C |
|
 |
| Glutamine, Gln, Q |
|
 |
| Methionine, Met, M |
|
 |
| Serine, Ser, S |
|
 |
| Threonine, Thr, T |
|
| back to top |
|
Hydrophobicity Index for Common Amino Acids |
The hydrophobicity index is a measure of the relative hydrophobicity, or how soluble an amino acid is in water. In a protein, hydrophobic amino acids are likely to be found in the interior, whereas hydrophilic amino acids are likely to be in contact with the aqueous environment.
The values in the table below are normalized so that the most hydrophobic residue is given a value of 100 relative to glycine, which is considered neutral (0 value). The scales were extrapolated to residues which are more hydrophilic than glycine. |
|
|
| back to top |