Chemical Synthesis
Fluorous
Product Catalog
Technology Spotlights
Key Resources
Drug Discovery
Materials Science
Stable Isotopes (ISOTEC)
 Fluorous Catalysts and Ligands
Fluorous Chemistry
Biphasic catalysis was the first application of fluorous chemistry in 1994. Since that time, fluorous biphasic catalytic methods have advanced rapidly, and a large number of fluorous catalysts and ligands (especially phosphines) are known. The defining feature of FBC is the use of a fluorous reaction solvent, and the technique is best viewed as a liquid phase catalyst immobilization method.
Fluorous Phosphines
Fluorous phosphines have been extensively used as metal ligands in fluorous biphasic catalysis. By varying the number and length of the fluorous chains, reaction and separation characteristics can be tuned. Fluorous phosphines and their oxides can be easily removed using either fluorous solid phase extraction or fluorous liquid-liquid extraction.
Triphenylphosphine
FTI Product: F017039 (07026)
Triphenylphosphine Structure Image
Fluorous triphenylphosphine, F017039, is often used in conjunction with the fluorous DIAD for traditional solution phase Mitsunobu chemistry and in biphasic catalysis.
Application Note
Triphenylphosphine, Ethylene Spacer
FTI Products: F026040 (19971), F034040 (16367)
Triphenylphosphine, Ethylene Spacer Structure Image
Triphenylphosphine, Branched
FTI Product: F026041 (50476)
Triphenylphosphine, Branched Structure Image
Triphenylphosphine, No Spacer
FTI Products: F039038 (12118), F051038 (07086)
Triphenylphosphine, No Spacer Structure Image
Triphenylphosphine, Ethylene Spacer
FFTI Product: F039042 (49317)
Triphenylphosphine, Ethylene Spacer Structure Image
Triphenylphosphine, Branched
FTI Product: F039043 (67301)
Triphenylphosphine, Branched Structure Image
Other Fluorous Phosphines