Sigma-Aldrich
Aldrich - Chemistry logo
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
ChemFilesTitle Vol. 2, No. 1
Products for Suzuki Coupling
Download PDF (1.03 MB)
Introduction / Boronic Acids / Boronic Acids Esters / Diboron Esters / CombiKits™
Prolines & Pyroglutamates / Transition-Metal Catalysts / Palladium Scavengers
FT-NMR Libraries on CD-ROMs / Large-Scale Suzuki Coupling / Reference Books / References
Introducing...
NEW Electronic FT-NMR Libraries from Aldrich!
The Aldrich FT-NMR Library with Spectral Viewer on CD-ROM is an electronic book that contains high-resolution 300MHz 1H and 75MHz 13C spectra for the more than 11,800 compounds found in the printed version of The Aldrich Library of 13C and 1H FT-NMR Spectra. A supplemental library is also available that includes spectra for 3,500 additional compounds not included in the original printed version. Order both the standard and the supplemental library for access to spectra on over 15,000 compounds.
Z541265a   
Click here to learn more about the Spectral Viewer.
Features:
  • Displays 1H and 13C spectra, integral, chemical structure, and technical data
  • Text Searching by Chemical Name, Aldrich Catalog Number, CAS Number, and Molecular Formula
  • Advanced data field searching on Chemical Category and physical properties such as Boiling Point, Melting Point, Density, Flash Point, Molecular Weight, and Refractive Index
  • Spectra processing using Zoom, Peak Integration, and Peak Labeling
  • Print spectra, structure, technical data, integral table and peak listing
  • Export spectra to a file (Bitmap, Metafile, or JPEG formats) or to the clipboard (Bitmap or Metafile formats)
  • Web link button provides additional technical data from the Sigma-Aldrich Website
Ideal for:
  • Teaching students NMR spectral interpretation
  • Referencing spectra for comparison studies and the identification of unknown compounds
  • Exporting spectra quickly for presentations and papers
Version Standard library
(11,800 compounds)
Cat. No.
Supplemental library
(3,500 compounds)
Cat. No.
Single user, commercial Z54,126-5 Z53,808-6
Single user, academic Z54,138-9 Z53,818-3
Network version (1-10 users) Z54,149-4 Z53,797-7
Demo program Z54,159-1
Minimum system requirements: Pentium® 200MHz, Windows® 95/98/ME/NT4-SP5/2000, CD-ROM drive, 600MB hard disk space, 64MB RAM. (PC ONLY)
Aldrich is a registered trademark of Sigma-Aldrich Co.
Chemical Abstracts is a registered trademark of the American Chemical Society.
Pentium is a registered trademark of Intel Corp. Windows is a registered trademark of Microsoft Corp.