Sigma-Aldrich
Aldrich - Chemistry logo
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Chemistry > Chemical Synthesis > Learning Center > ChemFiles > ChemFiles 2001-2003 > Vol 2 No 1 > Transition Metal Catalysts
ChemFilesTitle Vol. 2, No. 1
Products for Suzuki Coupling
Download PDF (1.03 MB)
Introduction / Boronic Acids / Boronic Acids Esters / Diboron Esters / CombiKits™
Prolines & Pyroglutamates / Transition-Metal Catalysts / Palladium Scavengers
FT-NMR Libraries on CD-ROMs / Large-Scale Suzuki Coupling / Reference Books / References
Transition-Metal Catalysts
A variety of transition-metal catalysts for the Suzuki coupling reaction are available from Aldrich. The majority of these catalysts are palladium- and nickel-based, typically utilizing phosphine-derived ligands.6-8
Cat. No. Product Information Unit Sizes
47,055-4 [1,1'-Bis(diphenylphosphino)ferrocene]dichloronickel(II), 97% 1g
37,967-0 [1,1'-Bis(diphenylphosphino)ferrocene]dichloropalladium(II), complex with CH2Cl2 1g; 5g
40,323-7 Dichlorobis(tricyclohexylphosphine)palladium(II), 95% 1g
45,304-8 Dichlorobis(tri-o-tolylphosphine)palladium(II), 97% 1g; 5g
20,867-1 Dichlorobis(triphenylphosphine)palladium(II), 98% 1g; 5g; 25g
41,274-0 Dichlorobis(triphenylphosphine)palladium(II), 99.99% 250mg; 1g
20,586-9 Palladium(II) acetate, 98% 1g; 2g; 10g
37,987-5 Palladium(II) acetate, 99.98% 1g; 5g
21,666-6 Tetrakis(triphenylphosphine)palladium(0), 99% 1g; 5g; 25g
32,877-4 Tris(dibenzylideneacetone)dipalladium(0) 500mg; 5g; 50g
36,631-5 Tris(dibenzylideneacetone)dipalladium(0)-chloroform adduct 250mg; 1g