Sigma-Aldrich
Aldrich - Chemistry logo
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
ChemFilesTitle Vol . 2, No. 4
Unnatural Amino Acids II
The latest Update on New Tools for Drug Discovery
Click here to download PDF version. (688 KB)
Introduction / Cyclic Amino Acids / Diamino Acids / ß-Amino Acids and Homo Amino Acids
Alanine Derivatives / Phenylalanine Boronic Acids / Proline and Pyroglutamine Derivatives
Other Amino Acid Building Blocks / Coupling Reagents / Preloaded Resins / Books
Coupling Reagents
Fluka is a global supplier of the highest quality coupling reagents, available from research to large-scale quantities. We focus on a broad assortment of versatile peptide coupling reagents, carbodiimides and additives for achieving fast, efficient, selective and racemization-free amidation in a wide variety of solvent systems.
Carbodiimides
The most popular in-situ condensing agents are the carbodiimides.[1-3] The reaction of a carboxylic acid with a carbodiimide is believed to involve a labile O-acylisourea (see figure).
carbodiimide_diagram
N,N4-dicyclohexylcarbodiimide (DCC) is extensively used in Boc/Bzl-peptide synthesis, because the dicyclohexylurea (DCU) by-product is easily removed from the reaction vessel in presence of trifluoroacetic acid during the Boc-deprotection protocol. In the Fmoc/t-Bu chemistry, diisopropylcarbodiimide gives rise to a more DMF-soluble urea by-product and is therefore highly recommended.4,5 N-(3-dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride (WSC, EDC) is widely used in solution phase, as it generates a urea by-product which can be easily removed from the reaction medium by extraction with water.6,7
Another important water-soluble carbodiimide is N-cyclohexyl-N'-(2-morpholinoethyl)carbodiimide-methyl-p-toluenesulfonate (CMC).8,9 A major drawback of the cabodiimide procedure is the dehydration of side-chain carboxamides of Asn- and Gln-residues to the corresponding nitriles. This problem is completely avoided when using carbodiimides in combination with additives like hydroxylamine derivatives, such as 1-hydroxybenzotriazole (HOBt)10 or 7-aza-1-hydroxybenzotriazole (HOAt).11-13 These additives lead to an efficient suppression of racemization and total exclusion of dehydration of carboxamide residues while generating highly active ester species. Carbodiimide-mediated activation is achieved without base catalysis otherwise responsible for considerable rates of racemization in sensitive Fmoc-Cys(Trt)-OH residue.14,15
Cat. No. Product Information Unit Sizes
36650 N,N'-Dicyclohexylcarbodiimide (DCC)
~99% C13H22N2 Mr 206.33 [538-75-0] 100g; 500g; 2.5kg
36651 N,N'-Dicyclohexylcarbodiimide solution, ~1 M in NMP 100ml
36652 N,N'-Dicyclohexylcarbodiimide solution, ~1 M in dichloromethane 100ml
38370 N,N'-Diisopropylcarbodiimide
>98.0% C7H14N2 Mr 126.20 [693-13-0] 25ml; 100ml; 500ml
34640 N,N'-Di-tert-butylcarbodiimide
>99.0% C9H18N2 Mr 154.26 [691-24-7]   5ml; 25ml
03449 N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride (WSC, EDC)
>99.0% C8H17N3•HCl Mr 191.70 [25952-53-8]   1g; 5g; 25g
39391 N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide (WSC, EDC)
>97.0% C8H17N3 Mr 155.24 [25952-53-8]   10ml; 50ml
29469 N-Cyclohexyl-N'-(2-morpholinoethyl)carbodiimide methyl-p-toluenesulfonate (CMC)
>99.0% C14H26N3O•C7H7O3S Mr 423.58 [2491-17-0]   5g; 25g
29470 N-Cyclohexyl-N'-(2-morpholinoethyl)carbodiimide methyl-p-toluenesulfonate (CMC)
>97.0%         5g; 25g
02541 2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline (EEDQ)
>99.0% C14H17NO3 Mr 247.30 [16357-59-8] structure 10g; 50g
21860 1,1'-Carbonyldiimidazole (CDI)
~97% C7H6N4O Mr 162.15 [530-62-1] structure 5g; 25g; 100g
21861 1,1'-Carbonyldi(1,2,4-triazole) (CDT)
~95% C5H4N6O Mr 164.13 [41864-22-6] structure 5g; 25g
15118 Bis(4-nitrophenyl) carbonate
>97.0% C13H8N2O7 Mr 304.21 [5070-13-3] structure 10g; 50g
23240 4-Nitrophenyl chloroformate
>97.0% C7H4ClNO4 Mr 201.57 [7693-46-1] structure 10g; 50g; 250g
43720 Di(N-succinimidyl) carbonate (DSC)
>97% C9H8N2O7 Mr 256.17 [74124-79-1] structure 5g; 25g
29224 4-Dimethylaminopyridine (DMAP)
>99.0% C7H10N2 Mr 122.17 [1122-58-3]   10g; 50g; 250g
39405 4-Dimethylaminopyridine (DMAP) >98.0% structure 10g; 50g; 250g
54804 1-Hydroxybenzotriazole hydrate (HOBt)
>99.0% C6H5N3O•aq Mr 135.13 [123333-53-9]   100g; 250g
54802 1-Hydroxybenzotriazole hydrate (HOBt)
>98.0% C7H10N2 Mr 122.17 [1122-58-3] structure 10g; 50g; 250g
54810 1-Hydroxybenzotriazole solution (HOBt) ~1 M in NMP 100ml; 500ml
12815 1-Hydroxybenzotriazole solution (HOBt), ≤0.1% water
~0.2 M in DMSO/NMP 50ml
37305 3,4-Dihydro-3-hydroxy-4-oxo-1,2,3-benzotriazine (Dhbt)
>98.0% C7H5N3O2 Mr 163.14 [28230-32-2] structure 10g; 50g; 250g
56480 N-Hydroxysuccinimide (HOSu)
>97.0% C4H5NO3 Mr 115.09 [6066-82-6] structure 25g; 100g; 1kg
56485 N-Hydroxysulfosuccinimide sodium salt (Sulfo-NHS)
>98.5% C4H4NNaO6S Mr 217.13 [106627-54-7] structure 250mg; 1g; 5g
56191 N-Hydroxyphthalimide
>98.0% C8H5NO3 Mr 163.13 [524-38-9] structure 100g; 500g
56055 N-Hydroxy-5-norbornene-2,3-dicarboxylic acid imide (HONB)
>98.0% C9H9NO3 Mr 179.18 [1122-58-3] structure 10g; 50g
63941 1-(2-Mesitylenesulfonyl)-3-nitro-1H-1,2,4-triazole (MSNT)
>98.0% C11H12N4O4S Mr 296.31 [74257-00-4] structure 1g; 5g
76740 2,3,4,5,6-Pentafluorophenol (Pfp-OH)
>99.0% C6HF5O Mr 184.07 [771-61-9] structure 5g; 25g
REFERENCES:
  1. Sheehan, J. C., Hess, G.P., J. Am. Chem. Soc., 1955, 77, 1067.
  2. Rich, D. H., Singh, J., in The Peptides: Analysis, Synthesis, Biology, (E. Gross, J. Meienhofer, eds), Vol. 1, Academic Press, New York 1979, 242.
  3. Beyermann, M. et al., Int. J. Pept. Protein Res., 1991, 37, 25.
  4. Sarantakis, D. et al., Biochem. Biophys. Res. Commun., 1976, 73, 336.
  5. Hudson, D. et al., in Peptide Chemistry, Proc. 23rd Japn. Pept. Symp., (Y. Kiso, ed.), Protein Research Foundation, Osaka, 1986, 4113.
  6. Sheehan, J. C., Ledis, S. L., J. Am. Chem. Soc., 1973, 95, 875.
  7. Sakakibara, S., Biopolymers (Pept. Sci.), 1995, 37, 17.
  8. Sheehan, J. C., Hlavka, J. J., J. Org. Chem., 1956, 21, 4395.
  9. Kunz, H., Angew, Chem., 1978, 90, 63.
  10. Kvnig, W. Geiger, R., Chem. Ber. 1970, 103, 788.
  11. Carpino, L. A., J. Am. Chem. Soc. 1993, 115, 4397.
  12. Carpino, L. A.et al., J. Chem. Soc., Chem. Commun., 1994, 201.
  13. Carpino, L. A. et al., Tetrahedron Lett.,1994, 35, 2279.
  14. Kaiser, T. et al., Tetrahedron Lett., 1996, 37, 1187.
  15. Meisenbach, M. et al., J. Chem. Soc., Chem. Commun., 1997, 849.