Sigma-Aldrich
Aldrich - Chemistry logo
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
ChemFilesTitle Vol . 2, No. 4
Unnatural Amino Acids II
The latest Update on New Tools for Drug Discovery
Click here to download PDF version. (688 KB)
Introduction / Cyclic Amino Acids / Diamino Acids / ß-Amino Acids and Homo Amino Acids
Alanine Derivatives / Phenylalanine Boronic Acids / Proline and Pyroglutamine Derivatives
Other Amino Acid Building Blocks / Coupling Reagents / Preloaded Resins / Books
Diamino Acids
Cat. No. Product Information Unit Sizes
32840 H-DL-Dap-OH dihydrochloride DL-2,4-Diaminobutyric acid dihydrochloride
>99.0% C4H10N2O2•2HCl Mr 191.06 [65427-54-5]   1g; 5g
76179 H-D-Dap-OH HCl D-2,3-Diaminopropionic acid monohydrochloride
>99.0% C3H8N2O2•HCl Mr 140.57 [6018-56-0]   1g; 5g
73031 Boc-Dap(Fmoc)-OH Na-Boc-Nß-Fmoc-L-2,3-diaminopropionic acid
>98.0% C23H26N2O6 Mr 426.46 [122235-70-5]   500mg
86283 Boc-Dap(Boc)-OH DCHA Na,Nß-Di-Boc-L-2,3-diaminopropionic acid Dicyclohexylamine salt
~97% C13H24N2O6•C12H23N Mr 485.66 [201472-68-6] structure 500mg; 2.5g
94572 Boc-Dap(Alloc)-OH DCHA Nß-Alloc-Na-Boc-L-2,3-diaminopropionic acid Dicyclohexylamine salt
>97.0% C12H20N2O6 Mr 288.30 [204197-28-4] structure 1g; 5g
76289 Boc-Dap(Z)-OH DCHA Na-Boc-Nß-Z-L-2,3-diaminopropionic acid
>98.0% C16H22N2O6•C12H23N Mr 519.68 [65710-58-9] structure 500mg; 2.5g
74242 Fmoc-Dap(Z)-OH Na-Fmoc-Nß-Z-L-2,3-diaminopropionic acid
~97% C26H24N2O6 Mr 460.48 [204316-36-9]   500mg
96414 Z-Dap(Fmoc)-OH Na-Z-Nß-Fmoc-L-2,3-diaminopropionic acid
>98.0% C26H24N2O6 Mr 460.48 [142855-80-9]   500mg
74391 Fmoc-Dap(Fmoc)-OH Na,Nß-di-Fmoc-L-2,3-diaminopropionic acid
>97.0% C33H28N2O6 Mr 548.59 [201473-90-7]   500mg
83041 Boc-Dab(Fmoc)-OH Ng-Fmoc-Na-Boc-L-2,4-diaminobutyric acid
>98.0% C24H28N2O6 Mr 440.49 [117106-21-5] structure 250mg; 1g
81717 Boc-Dab(Alloc)-OH DCHA Ng-Alloc-Na-Boc-L-2,4-diaminobutyric acid Dicyclohexylamine salt
  C13H22N2O6•C12H23N Mr 483.65 [327156-92-3]   1g; 5g
88645 Fmoc-Dab(Alloc)-OH Ng-Alloc-Na-Fmoc-L-2,4-diaminobutyric acid
>98.0% C23H24N2O6 Mr 424.45 [204316-32-5] structure 1g; 5g