Chemical Synthesis
Product Catalog
Technology Spotlights
Key Resources
New Products Archive
Aldrichimica Acta
ChemFiles
ChemNews
Cheminars
ChemBlogs
Technical Bulletins
ACS Awards
Drug Discovery
Materials Science
Stable Isotopes (ISOTEC)
Vol. 2, No. 6
Functionalized SILICA GELS for Organic Synthesis
Download PDF (805 KB)
Introduction / Quality Control / Organic Synthesis / Scavengers
Supported Reagents / Linker Resins / Kits / References
Introduction
Sigma-Aldrich and SiliCycle Inc. have partnered to offer you a wide-range of functionalized silica gels for organic synthesis. SiliCycle Inc. manufacture these novel products to combine with the 700+ resins that Sigma-Aldrich now offers to help your synthetic needs.
These novel silica gel products offer numerous advantages over the well-known gel-type resins.
Advantages of the Inert Support
  • Solvent independent
    • Rigid porous structure
      • No swelling
      • Fast kinetics
      • Can be used in large-scale purification (discovery to process)
    • No low molecular weight residues
      • High purity
  • No non-specific binding
    • High Yield
  • High density of functional groups
    • Small volume of silica gel required
This ChemFile details the complete range of functionalized silica gels and their applications. All of these products are available in pack sizes of 5 grams to multi-kilos.