Vol. 2, No. 7
Eckert's Cartridges for Safe Phosgenations
Download PDF version (448KB)
Introduction /
Applications /
Phosgene and Substitutes /
Instructions for Use /
References
What if phosgene reagents were available off the shelf in your
lab? What if you could obtain them without any transport and storage
concerns? Or use them without complicated equipment, or worrying
about handling them due to the safety hazards commonly associated
with phosgene?
Impossible? NOT ANYMORE! Sigma-Aldrich now offers Eckerts
Cartridges for Safe Phosgenation (U. S. Patent 6,399,822
B1) to our customers worldwide. Based on the catalytic formation
of phosgene, the cartridge contains a unique, safe precursor capable
of generating this versatile reagent in quantities of 20 mmol and
50 mmol.
Eckerts Cartridges for Safe Phosgenation offer many
benefits:
- More versatile chemistry through phosgene, when other reagents offer poor results
- Fast reactions under mild conditions
- High conversion rates and yields
- Simple workup of reactions to obtain pure products
- Safe transport and storage
- Safe and easy handling through premeasured, commonly used quantities
- Time and cost efficiency
- Solvent-free reagent
- Less waste and easy disposal of used cartridges
These advantages of Eckerts cartridges provide easy access
to the world of phosgene chemistry.
Products
| Starter
Kit:
Contains one Cartridge for Safe Phosgene
Generation, 0.02mol (#51,975-8), one gastight adapter with
hose, one dosimeter badge + paper, and instructions |
1 kit |
51,978-2
|
|
Replacement Cartridges: |
| Cartridge for Safe Phosgene Generation, 0.02mol |
1 pk. 5 pk.
|
51,975-8
|
| Cartridge for Safe Phosgene
Generation, 0.05mol |
1 pk. 5 pk.
|
51,976-6