Vol . 2, 2002 Supplement
Grubbs' Catalysts & MacMillan's OrganoCatalysts
Download PDF (641 KB)
Grubbs' Catalysts /
MacMillan's OrganoCatalysts
Verkade's Superbases /
Metal Catalysts and Ligands /
References
|
Grubbs Catalysts
In todays fast-paced environment, ready access to leading
edge developments in catalyst technology by the modern synthetic
chemist is a defining competitive advantage. Aldrich is pleased
to announce a partnership with Materia, Inc. to offer research quantities
of novel organic and organometallic catalysts from Materia's technology
portfolio. Materia's olefin metathesis and OrganoCatalyst
technologies enable the production of fine chemicals and materials
in fewer steps, higher yields, and with fewer byproducts than traditional
routes.
|
Ruthenium-Based Metathesis Catalysts
|
Olefin metathesis is a synthetically powerful transformation
in which a net exchange of olefin substituents occurs. The
reaction between substrate and catalyst proceeds via the reversible
formation of a metallacyclobutane intermediate. A significant
evolution in the development of olefin metathesis catalysts
involves the use of ruthenium-based catalysts discovered in
the laboratories of Professor Robert Grubbs at Caltech. The
synthetic utility of ruthenium-based catalysts is derived
from their ability to orchestrate additional metathetical
transformations, including Ring-Closing Metathesis (RCM),
Ring-Opening Metathesis Polymerization (ROMP), and Acyclic
Diene Metathesis Polymerization (ADMET) (Figure 1).
These transformations enable the production of novel compounds
and high-performance materials for the pharmaceutical and
materials science markets.
|
 |
Pharmaceutical Applications
The application of ruthenium-based olefin metathesis technology
to the manufacture of pharmaceutical intermediates is boundless.
Perhaps most strikingly, Ring-Closing Metathesis (RCM) enables the
efficient production of complex ring systems from simple acyclic
precursors using Grubbs Catalyst, first generation (1)
(Scheme 1).1
RCM using 1 has also been utilized in a key step of the total synthesis
of Mevinolin (Scheme 2),2 a compound shown to
reduce serum cholesterol levels, and in the synthesis of Ambruticin
(Scheme 3),3 an orally active antifungal agent.
A further example illustrates the application of metathesis reactions
to peptidomimetics. RCM on acyclic peptides bearing olefin functionalities
results in the formation of cyclic species. The olefin generated
by metathesis in this case replaces the disulfide linkages found
in natural and synthetic peptides. The resulting carbocyclic substructure
has a reduced conformational space that often results in an increased
affinity for biological receptors, and provides dramatically improved
redox stability (Scheme 4).4-7
RCM has also been used in the synthesis of a-substituted and a,a-disubstituted
amino acids.8 A comparison of the product yield between
catalyst 1 and Grubbs Catalyst, second generation
(2) is given in Table 1. Calalyst 2 is more
active and exhibits excellent functional group tolerance and selectivity.
Catalyst concentration also influences overall yields, as demonstrated
in the RCM of an ene-ynamide system (Table 2) using Grubbs
Catalyst, second generation.9
Table 1:
 |
n |
Catalyst |
Yield (%) |
| 1 |
1 |
92 |
| 2 |
1 |
74 |
| 2 |
2 |
87 |
| 3 |
1 |
57 |
| 3 |
2 |
83 |
Table 2:
 |
| Run |
Substrate |
(mol %) |
Conditions |
Time(h) |
Yielda
(%) |
| 1 |
(R=H) |
5 |
toluene/80 °C |
20 |
22 |
| 2 |
|
5 |
toluene/60 °C |
20 |
19 (20) |
| 3 |
|
5 |
CH2CI2/reflux |
27 |
36 (38) |
| 4 |
|
10 |
CH2CI2/reflux |
6 |
85 |
| 5 |
|
10 |
CH2CI2/reflux |
5 |
82 |
| 6 |
(R=Me) |
10 |
CH2CI2/reflux |
6 |
61 |
a The yield in parenthesis
is that of the recovered starting material. The reaction was carried
out under argon.
Materials Applications
Ruthenium-based metathesis catalysts have shown remarkable utility
in the production of fine chemicals on an industrial scale by
RCM and Cross Metathesis (CM) reactions. ROMP and ADMET have been
used extensively in the field of materials science to produce
polymers with unique properties. One such example is the ROMP
of dicyclopentadiene (DCPD) to give resins that exhibit remarkable
impact and corrosion resistance. DCPD resin is an excellent base
resin for a variety of composite products used in areas such as
the defense/aerospace industry, sports and recreation, marine,
ballistics (Figure 2), and microelectronics.
RCM and Cross Metathesis (CM) can be combined to produce a wide
variety of terminally functionalized oligomers or polymers, commonly
referred to as telechelic materials (Scheme 5).10,11
These materials are notoriously difficult or impossible to prepare
in high yield and selectivity by standard synthetic methods, but
have several important and growing applications including the
production of multi-block polymers and thermoplastic polyurethanes.
Figure 2
|
|
Ferroelectric liquid crystals (FLC) have been prepared via ADMET
polymerization using a derivative of Grubbs Catalyst, first
generation. The predominant species is the acyclic oligomer with
little competition from the RCM cyclic species (Scheme 6).12
Hoveyda-Grubbs' Catalyst
Invented at Boston College, the Hoveyda-Grubbs Catalyst
(3) shows efficiencies similar to those of Grubbs Catalyst
second generation, but with a different substrate specificity.
The recyclable13 catalyst is unique in catalyzing RCM,
ROMP, and CM reactions with highly electron-deficient substrates
(Scheme 7).14
 |
 |
 |
09587
Grubbs Catalyst,
First Generation
Benzylidenebis-
(tricyclohexylphosphine)-
dichlororuthenium
250mg; 1g; 5g |
56,974-7
Grubbs Catalyst,
Second Generation
1,3-(Bis(mesityl)-2-
imidazolidinylidene)dichloro-
(phenylmethylene)(tricyclohexyl-
phosphine)ruthenium
500mg; 2g |
56,975-5
Hoveyda-Grubbs
Catalyst
1,3-(Bis(mesityl)-2-
imidazolidinylidene)dichloro-
(o-isopropoxyphenylmethylene)
ruthenium
250mg; 1g; 5g |
|