|
ChemFiles
OrganoCatalysts
|
|
|
| |
Vol . 2, 2002 Supplement Grubbs' Catalysts & MacMillan's OrganoCatalysts™ Download PDF (641 KB) Grubbs' Catalysts / MacMillan's OrganoCatalysts™ Verkade's Superbases / Metal Catalysts and Ligands / References
MacMillan's OrganoCatalysts™
The vast majority of useful catalytic transformations involve inorganic or organometallic complexes as catalysts. Examples of catalysis by organic molecules can be found, but these examples are rare and have no general utility. OrganoCatalysis™ is a breakthrough technology, invented by Professor David MacMillan at Caltech, in which small organic molecules are designed and constructed to serve as general catalysts for asymmetric transformations of other substrates.
OrganoCatalysts™ have been successfully developed for several important reactions including the asymmetric variants of Friedel-Crafts alkylations,15 1,4 conjugate additions, Diels–Alder reactions,16 and 1,3 dipolar cycloadditions.17 These transformations enable an extremely broad range of chiral intermediates to be readily assembled (Scheme 8).
An interesting example employing RCM via Grubbs’ Catalyst in tandem with OrganoCatalysts™ is the three-step enantioselective synthesis of (-)-Ketorolac (Scheme 9). Ketorolac is currently manufactured in racemic form in a ten-step process. Studies of the pure (S)-enantiomer show enhanced biological activity and lower toxicity versus the racemic mixture.
OrganoCatalysts and OrganoCatalysis are trademarks of Materia, Inc.
 |
 |
56,976-3
(5S)-2,2,3-Trimethyl-5-phenylmethyl-4-imidazolidinone monohydrochloride
2g |
56,977-1
(2S,5S)-2-(1’,1’-Dimethylethyl)-3-methyl-5- phenylmethyl-4-imidazolidinone
2g |