Chemical Synthesis
Product Catalog
Technology Spotlights
Key Resources
New Products Archive
Aldrichimica Acta
ChemFiles
ChemNews
Cheminars
ChemBlogs
Technical Bulletins
ACS Awards
Drug Discovery
Materials Science
Stable Isotopes (ISOTEC)
Vol. 3, No. 2
Products For
Biomolecular NMR
Download PDF version (1.49 MB)
Page 1: Intro / Labeled Products for Cell Free Synthesis / Products for Peptide Synthesis / Products for Minimal Media
Page 2: Labeled Complex Growth Media / Buffers and Reagents / a-Keto Acids / Labeled Nucleotides
Page 3: Custom Synthesis / Aldrich Spectral Viewer / NEW - ISOTEC 2003-2004 Capabilities Catalog / ISOTEC Quality
Volume 3 Number 2
Products for Biomolecular NMR
Modern multiple resonance and multi-dimensonal Biomolecular NMR experiments require isotopic enrichment of the proteins, RNA and DNA, to achieve sufficient sensitivity and resolution. The biomolecules are typically enriched by biosynthesis. ISOTEC™ offers an extensive line of stable isotope labeled products for Biomolecular NMR applications.
Stable Isotopes in Cell-Free Protein Synthesis
Munehiro Teshima-Scientist-ISOTEC™
Proteomics is an emerging field of post genome research, which examines the enormous amounts of information stored in the genomes of living organisms.1 Biomolecular NMR and stable isotopes are important tools used to study the structure and function of proteins in structural genomics research. Most researchers use the protein expression systems of microorganisms or mammalian cells and stable isotope labeled (2H, 13C, 15N) D-glucose, ammonium salts, amino acids and/or complex growth media to produce proteins uniformly labeled with stable isotopes.
However, the protein expression system using living cells has limitations.
  • Many expressed proteins are insoluble and aggregate in inclusion bodies.
  • Intercellular proteases in the host cells may digest the proteins.
  • Some proteins cannot be produced in living cells because of their toxicity.
  • The amino acid metabolic system in the host cells can cause isotope dilution and diffusion for amino acids
  • with selective stable isotope labeling.
  • It may be difficult to grow organisms in deuterium labeled media.
  • Procaryote cannot glycosylate proteins.
These problems associated with living cells may be solved by the cell-free protein synthesis system. An additional benefit is that the cell-free protein synthesis system is suitable for automation and high throughput protein synthesis.
The basic idea of the cell-free protein synthesis system is to extract only the protein synthesis system (translation and / or transcription system) from living cells such as Escherichia Coli, rabbit reticulocyte or wheat germ, and allow the extract to synthesize the proteins in vitro from amino acids and specific DNA or mRNA used as a template.
The cell-free protein synthesis system is not a new technology, but the yield of the conventional batch type system used in the past was so low that radioisotopes were the only method sensitive enough for detection. In 1988, Dr. Spirin et al. developed a continuous flow cell-free protein synthesis system. They used an ultra-filtration system and succeeded in producing a couple of hundred mg of protein per ml of reaction mixture.2
Many ideas for the improvement of the continuous flow system such as using dialysis and condensation of the extract were incorporated by many researchers.3-7 In addition to these improvements, Dr. Yokoyama et al. optimized the reaction conditions and established a system to produce 6 mg of protein per ml of reaction mixture, and they applied this system to produce 13C, 15N-labeled proteins for Biomolecular NMR spectroscopy.8 Also, they succeeded in developing a site directed stable isotope labeling method for a protein by using a cell-free protein synthesis system.9
References:
  1. http://www.gsc.riken.go.jp/e/group/protgrE.html.
  2. Sprin, A. S., et al., Science, 1988, 1162-1164, 242.
  3. Kim, D. M., et al., Biotechnol. Prog., 1996, 645-649, 12.
  4. Davis, J., et al., Promega Notes Mag. 1996, 14-18, 56.
  5. Nakano, H., et al., Biosci. Biotechnol. Biochem. 1994, 631-634, 58.
  6. Kim, D. M., et al., Eur. J. Biochem. 1996, 881-886, 239.
  7. Nakano, H., et al., J. Biotechnol. 1996, 275-282, 46.
  8. Yokoyama, S., et al., FEBS Lett. 1999, 15-19, 442.
  9. Yokoyama, S., et al., J. Biomol. NMR 1998, 295-306, 11.
Top
Labeled Products for Cell Free Synthesis
Fully labeled 15N Amino Acids
Cat. No. Product Name Atom %
33,212-7 L-Alanine-15N 98
60,011-3 L-Arginine-15N4 HCl 98
48,591-8 L-Alanine-15N 98
33,213-5 L-Asparagine-15N2•H2O 98
60,912-9 L-Aspartic-15N Acid 98
33,214-3 L-Glutamic-15N Acid 98
49,003-2 L-Glutamine-15N2 98
29,929-4 Glycine-15N 98
57,436-8 L-Histidine-15N3 95
33,215-1 L-4-Hydroxyphenlyalanine-15N (L-Tyrosine) 98
60,901-3 L-Isoleucine-15N 98
34,096-0 L-Leucine-15N 98
60,902-1 L-Lysine-15N2 HCl 98
60,924-2 L-Methionine-15N 98
49,010-5 L-Phenylalanine-15N 98
60,899-8 L-Proline-15N 98
60,900-5 L-Serine-15N 98
60,909-9 L-Threonine-15N 98
57,460-0 L-Tryptophan-15N2 95
49,017-2 L-Valine-15N 98
Fully labeled 13C, 15N Amino Acids
Cat. No. Product Name Atom %
48,988-3 L-Alanine-13C3, 15N 98 13C; 98 15N
60,803-3 L-Arginine-13C6, 15N4 HCl 98 13C; 98 15N
60,815-7 L-Asparagine-13C4, 15N2•H2O 98 13C; 98 15N
60,783-5 L-Aspartic-13C4, 15N Acid 98 13C; 98 15N
60,785-1 L-Glutamic-13C5, 15N Acid 98 13C; 98 15N
60,798-3 L-Glutamine-13C5, 15N2 98 13C; 98 15N
48,952-2 Glycine-13C2, 15N 98 13C; 98 15N
60,800-9 L-Histidine-13C6, 15N3 97 13C; 95 15N
60,799-1 L-4-Hydroxyphenlyalanine-13C9, 15N (L-Tyrosine) 99 13C; 98 15N
60,809-2 L-Isoleucine-13C6, 15N 98 13C; 98 15N
60,806-8 L-Leucine-13C6, 15N 98 13C; 98 15N
60,804-1 L-Lysine-13C6, 15N2 HCl 98 13C; 98 15N
60,810-6 L-Methionine-13C5, 15N 98 13C; 98 15N
60,801-7 L-Phenylalanine-13C9, 15N 98 13C; 98 15N
60,811-4 L-Proline-13C5, 15N 98 13C; 98 15N
60,813-0 L-Serine-13C3, 15N 98 13C; 98 15N
60,777-0 L-Threonine-13C4, 15N 98 13C; 98 15N
57,459-7 L-Tryptophan-13C11, 15N2 97 13C; 95 15N
60,014-8 L-Valine-13C5, 15N 98 13C; 98 15N
Top
Products for Peptide Synthesis
(N-FMOC and N-t-BOC Derivatives)
Cat. No. Product Name Atom %
48,583-7 L-Alanine-13C3,15N,N-t-BOC 98 13C; 98 15N
48,990-5 L-Alanine-15N,N-FMOC 98
48,991-3 L-Alanine-15N,N-t-BOC 98
57,989-0 L-Asparagine-15N2,a-N-FMOC 98
49,290-6 L-Aspartic-15N Acid,N-FMOC 98
57,795-2 L-Aspartic-15N Acid,N-FMOC,a-O-t-butyl 98
58,879-2 L-Aspartic-15N Acid,N-t-BOC 98
58,840-7 L-Glutamic Acid-13C5,15N,N-t-BOC,a-O-Benzyl Ester (97%CP) 9813C; 9815N
49,000-8 L-Glutamic-15N Acid,N-FMOC 98
60,915-3 L-Glutamic-15N Acid,N-FMOC,g-O-t-Butyl Ester 98
58,769-9 L-Glutamic-15N Acid,N-t-BOC 98
58,770-2 L-Glutamine-15N2,a-N-t-BOC 98
48,953-0 Glycine-13C2,15N,N-FMOC 9813C; 9815N
58,773-7 Glycine-13C2,15N,N-t-BOC 9813C; 9815N
48,575-6 Glycine-15N,N-FMOC 98
48,670-1 Glycine-15N,N-t-BOC 98
59,109-2 L-4-Hydroxyphenylalanine-15N,N-t-BOC(L-Tyrosine) 98
59,722-8 L-Isoleucine-13C6,15N,N-FMOC 9813C; 9815N
57,862-2 L-Isoleucine-15N,N-FMOC 98
48,595-0 L-Leucine-15N,N-FMOC 98
49,293-0 L-Leucine-15N,N-t-BOC 7 H2O 98
57,796-0 L-Lysine-15N2,a-N-FMOC,e-N-t-BOC 98
60,919-6 L-Methionine-15N,N-FMOC 98
60,907-2 L-Phenylalanine-15N,N-FMOC 98
48,683-3 L-Phenylalanine-15N,N-t-BOC 98
58,951-9 L-Proline-15N,N-FMOC 98
60,914-5 L-Serine-15N,N-FMOC,O-t-Butyl 98
48,600-0 L-Valine-15N,N-FMOC 98
48,601-9 L-Valine-15N,N-t-BOC 98
Top
Products for Minimal Media
Sugars
Cat. No. Product Name Atom %
38,937-4 D-Glucose-13C6 99
55,215-1 D-Glucose-13C6,C-d7 99 13C; 97-99 D
55,200-3 D-Glucose-C-d7 97-99
61,633-8 D-Glucose-d12 97-99
15N Salts
Cat. No. Product Name Atom %
29,925-1 Ammonium-15N Chloride 98
36,650-1 Ammonium-15N, d4 Chloride 98 15N; 98 D
48,801-1 Ammonium-15N Hydroxide (~3N aqueous soln.) 98
29,928-6 Ammonium-15N2 Sulfate 98
59,399-0 Ammonium-15N2, d8 Sulfate 99 15N; 98 D