Sigma-Aldrich
Aldrich - Chemistry logo
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
ChemFilesTitle Vol. 3, No. 3
Reagents for Suzuki Coupling
Download PDF
Introduction / Boronic acids / Boronic acid Esters / Diboron Esters / Potassium Trifluoroborates
Pd-Catalysts with Perfluoroalkyl Ponytails / OSVB, Volume 3: Suzuki Coupling
Boronic acids
Most boronic acids readily undergo dehydration reactions to give a cyclic (trimer) anhydride. Our selection of boronic acids may contain varying amounts of this cyclic anhydride. Fortunately, the acid and the anhydride work equally well in the Suzuki coupling reactions; thus, the two forms are generally regarded as equivalent.
<
Methylboronic acid trans-2-Bromomethylvinylboronic acid
16,533-6 CH5BO2 1g; 5g 55,656-4 C3H6BBrO2 1g; 5g
165336
new_bullet
  5565641
  trans-2-Chloromethylvinylboronic acid   cis-Propenylboronic acid
55,659-9 C3H6BClO2 1g; 5g 57,217-9 C3H7BO2 1g; 5g
5565991
572179
  trans-Propenylboronic acid 5-Bromothiophene-2-boronic acid
57,663-8 C3H7BO2 1g; 5g 55,768-4 C4H4BBrO2S 5g
576638
557684
5-Chloro-2-thiopheneboronic acid 2-Thiopheneboronic acid
49,993-5 C4H4BClO2S 5g 43,683-6 C4H5BO2S 1g; 5g
4999351
4368361
3-Thiopheneboronic acid 2-Furanboronic acid
43,684-4 C4H5BO2S 1g; 5g 46,491-0 C4H5BO3 1g; 10g
4368441
4649101
3-Furanboronic acid 2,5-Thiophenediboronic acid
51,216-8 C4H5BO3 1g 47,031-7 C4H6B2O4S 5g
5121681
4703171
Butylboronic acid (2-Methylpropyl)boronic acid
16,324-4 C4H11BO2 1g; 5g; 25g 34,622-5 C4H11BO2 1g; 5g
1632441
346225
2-Formyl-3-thiopheneboronic acid 3-Formyl-2-thiopheneboronic acid
49,990-0 C5H5BO3S 1g; 5g 49,991-9 C5H5BO3S 1g; 5g
4999001
4999191
5-Formyl-2-thiopheneboronic acid 4-Methyl-3-thiopheneboronic acid
51,405-5 C5H5BO3S 1g; 5g 54,239-3 C5H7BO2S 1g
5140551
542393
5-Methyl-2-thiopheneboronic acid   (E)-5-Chloro-1-penteneboronic acid
51,219-2 C5H7BO2S 1g; 5g 56,279-3 C5H10BClO2 1g; 10g
5121921
5627931
  1-Pentenylboronic acid Pentafluorophenylboronic acid
57,845-2 C5H11BO2 1g; 5g 46,509-7 C6H2BF5O2 5g
578452
4650971
2,3,4-Trifluorophenylboronic acid 2,3,6-Trifluorophenylboronic acid
52,408-5 C6H4BF3O2 5g 52,409-3 C6H4BF3O2 5g
524085
5240931
2,4,6-Trifluorophenylboronic acid 3,4,5-Trifluorophenylboronic acid
52,410-7 C6H4BF3O2 1g; 5g 52,470-0 C6H4BF3O2 1g
5241071
524700
3,5-Dibromophenylboronic acid 2-Chloro-6-fluorophenylboronic acid
49,950-1 C6H5BBr2O2 1g; 5g 56,607-1 C6H5BClFO2 1g; 5g
4995011
566071
3-Chloro-4-fluorophenylboronic acid 2,3-Dichlorophenylboronic acid
51,223-0 C6H5BClFO2 5g 51,404-7 C6H5BCl2O2 5g; 25g
5122301
5140471
2,4-Dichlorophenylboronic acid 2,5-Dichlorophenylboronic acid
52,138-8 C6H5BCl2O2 5g 51,231-1 C6H5BCl2O2 5g
5213881
5123111
3,4-Dichlorophenylboronic acid 3,5-Dichlorophenylboronic acid
47,191-7 C6H5BCl2O2 5g; 25g 44,520-7 C6H5BCl2O2 5g; 25g
4719171
4452071
2,3-Difluorophenylboronic acid 2,4-Difluorophenylboronic acid
51,403-9 C6H5BF2O2 5g; 25g 46,507-0 C6H5BF2O2 5g; 25g
5140391
4650701
2,5-Difluorophenylboronic acid 2,6-Difluorophenylboronic acid
51,402-0 C6H5BF2O2 5g; 25g 47,079-1 C6H5BF2O2 1g; 10g
514020
4707911
3,4-Difluorophenylboronic acid 3,5-Difluorophenylboronic acid
46,508-9 C6H5BF2O2 5g; 25g 47,192-5 C6H5BF2O2 5g; 25g
4650891
4719251
2-Bromophenylboronic acid 3-Bromophenylboronic acid
47,380-4 C6H6BBrO2 5g; 25g 44,162-7 C6H6BBrO2 1g; 5g
4738041
4416271
4-Bromophenylboronic acid 2-Chlorophenylboronic acid
B7,595-6 C6H6BBrO2 1g; 5g 44,521-5 C6H6BClO2 1g; 5g
B759561
4452151
3-Chlorophenylboronic acid 4-Chlorophenylboronic acid
41,752-1 C6H6BClO2 1g; 10g 41,754-8 C6H6BClO2 1g; 10g; 25g
4175211
4175481
2-Fluorophenylboronic acid 3-Fluorophenylboronic acid
44,522-3 C6H6BFO2 1g; 10g 44,164-3 C6H6BFO2 1g; 10g
4452231
4416431
4-Fluorophenylboronic acid 3-Iodophenylboronic acid
41,755-6 C6H6BFO2 1g; 5g; 25g 44,167-8 C6H6BIO2 5g; 25g
4175561
4416781
4-Iodophenylboronic acid 3-Nitrophenylboronic acid
47,193-3 C6H6BIO2 5g; 25g 32,510-4 C6H6BNO4 1g; 5g
471933
3251041
Phenylboronic acid 4-Mercaptophenylboronic acid
P2,000-9 C6H7BO2 10g; 50g; 250g 52,401-8 C6H7BO2S 1g; 5g
P20009
5240181
3-Hydroxyphenylboronic acid 4-Hydroxyphenylboronic acid
52,396-8 C6H7BO3 1g; 10g 52,397-6 C6H7BO3 1g; 5g
5239681
523976
5-Acetyl-2-thiopheneboronic acid 1,4-Phenylenebisboronic acid
49,992-7 C6H7BO3S 5g; 25g 41,713-0 C6H8B2O4 5g; 25g
4999271
417130
3-Aminophenylboronic acid hydrochloride 3-Aminophenylboronic acid monohydrate
41,070-5 C6H9BClNO2 1g; 5g 28,751-2 C6H10BNO3 1g; 5g
4107051
2875121
3-Aminophenylboronic acid hemisulfate Cyclohexylboronic acid
A7,175-1 C6H10BNO6S 5g; 25g 55,658-0 C6H13BO2 5g; 10g
A71751
5565801
  trans-1-hexen-1-ylboronic acid   trans-2-tert-butylvinylboronic acid
52,101-9 C6H13BO2 1g 55,655-6 C6H13BO2 1g; 5g
521019
5565561
2,3-Difluoro-4-formylphenylboronic acid 2-Fluoro-5-(trifluoromethyl)phenyl-boronic acid
57,137-7 C7H5BF2O3 1g; 5g 55,818-4 C7H5BF4O2 1g
571377
558184
2-(Trifluoromethyl)phenylboronic acid 3-(Trifluoromethyl)phenylboronic acid
44,519-3 C7H6BF3O2 1g; 10g 43,203-2 C7H6BF3O2 1g; 5g
445193
4320321
4-(Trifluoromethyl)phenylboronic acid 3-(Trifluoromethoxy)phenylboronic acid
43,932-0 C7H6BF3O2 1g; 5g 51,012-2 C7H6BF3O3 5g
4393201
510122
4-(Trifluoromethoxy)phenylboronic acid 2-Cyanophenylboronic acid
51,013-0 C7H6BF3O3 5g 52,139-6 C7H6BNO2 1g; 5g
510130
521396
3-Cyanophenylboronic acid 4-Cyanophenylboronic acid
51,301-6 C7H6BNO2 1g; 5g 52,141-8 C7H6BNO2 1g; 10g
5130161
5214181
2-Chloro-6-fluoro-3-methylphenyl-boronic acid 2-Chloro-6-fluoro-5-methylphenyl-boronic acid
55,725-0 C7H7BClFO2 5g 55,726-9 C7H7BClFO2 5g
557250
557269
2-Formylphenylboronic acid 3-Formylphenylboronic acid
43,195-8 C7H7BO3 1g; 5g 44,165-1 C7H7BO3 1g; 5g; 25g
4319581
4416511
4-Formylphenylboronic acid 3-Carboxyphenylboronic acid
43,196-6 C7H7BO3 1g; 5g; 25g 45,676-4 C7H7BO4 1g; 10g
4319661
456764
4-Carboxyphenylboronic acid 3,4-(Methylenedioxy)phenylboronic acid
45,677-2 C7H7BO4 1g; 10g 49,999-4 C7H7BO4 1g; 5g
4567721
499994