Sigma-Aldrich
Aldrich - Chemistry logo
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
ChemFilesTitle Vol. 3, No. 3
Reagents for Suzuki Coupling
Download PDF
Introduction / Boronic acids / Boronic acid Esters / Diboron Esters / Potassium Trifluoroborates
Pd-Catalysts with Perfluoroalkyl Ponytails / OSVB, Volume 3: Suzuki Coupling
Diboron Esters
The following diboron esters are versatile reagents that couple with organic triflates and halides to give the corresponding boronic esters, which are readily converted to arylboronic acids. This route and the subsequent Suzuki coupling reaction can be run under mild conditions, thus permitting the use of cyano-, ester-, carbonyl-, and nitro-substituted aryl rings. The wide variety of arylboronic acids available via these diboron esters also makes this class of compounds suitable for solid-phase combinatorial studies.
Bis(neopentyl glycolato)diboron Bis(catecholato)diboron
51,880-8 C10H20B2O4 1g; 5g 47,328-6 C12H8B2O4 1g; 5g
5188081
4732861
Bis(pinacolato)diboron Bis(hexylene glycolato)diboron
47,329-4 C12H24B2O4 1g; 5g; 25g; 100g 52,568-5 C12H24B2O4 1g; 5g
4732941
5256851
Bis(diethyl-D-tartrate glycolato)diboron Bis(diethyl-L-tartrate glycolato)diboron
52,715-7 C16H24B2O12 1g; 5g 52,716-5 C16H24B2O12 1g; 5g
527157
5271651
Bis(N,N,N',N'-tetramethyl-D-tartaramide glycolato)diboron Bis(N,N,N',N'-tetramethyl-L-tartaramide glycolato)diboron
52,720-3 C16H28B2N4O8 1g; 5g 52,723-8 C16H28B2N4O8 1g; 5g
527203
527238
Bis[(+)-pinanediolato]diboron Bis[(-)-pinanediolato]diboron
52,713-0 C20H32B2O4 1g; 5g 52,714-9 C20H32B2O4 1g; 5g
5271301
5271491
Bis(diisopropyl-D-tartrate glycolato)diboron Bis(diisopropyl-L-tartrate glycolato)diboron
52,717-3 C20H32B2O12 1g; 5g 52,718-1 C20H32B2O12 1g; 5g
527173
527181
pinacolatoAD