Sigma-Aldrich
Aldrich - Chemistry logo
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Chemistry > Chemical Synthesis > Learning Center > ChemFiles > ChemFiles 2001-2003 > Vol 3 No 3 > Potassium Trifluoroborates
ChemFilesTitle Vol. 3, No. 3
Reagents for Suzuki Coupling
Download PDF
Introduction / Boronic acids / Boronic acid Esters / Diboron Esters / Potassium Trifluoroborates
Pd-Catalysts with Perfluoroalkyl Ponytails / OSVB, Volume 3: Suzuki Coupling
Potassium Trifluoroborates
Alkyl, aryl, and alkenyl trifluoroborate salts are compelling alternatives to boronic acids in Suzuki coupling and in rhodium catalyzed carbon-carbon bond forming reactions. These salts are air and water stable, and in many cases eliminate the need to add either additional ligands or base for cross coupling. Whereas the boronic acids readily form cyclic anhydrides, these new salts do not form unwanted side products. In many cases, the trifluoroborate salt is more efficient and tolerant to functional groups than the corresponding boronic acids.
Potassium 3-thiophenetrifluoroborate Potassium 4-bromophenyltrifluoro-borate
57,157-1 C4H3BF3KS 1g; 5g 57,154-7 C6H4BBrF3K 1g; 5g
571571
571547
Potassium 2-bromophenyltrifluoro-borate Potassium 3-nitrophenyltrifluoro-borate
57,610-7 C6H4BBrF3K 1g; 5g 57,159-8 C6H4BF3KNO2 1g; 5g
576107
571598
Potassium phenyltrifluoroborate Potassium 4-(trifluoromethyl)phenyltri-fluoroborate
56,395-1 C6H5BF3K 1g; 5g 57,613-1 C7H4BF6K 1g; 5g
563951
576131
Potassium 4-formylphenyltrifluoro-borate Potassium benzyltrifluoroborate
57,609-3 C7H5BF3KO 1g; 5g 56,305-6 C7H7BF3K 1g; 5g
576093
563056
Potassium o-tolyltrifluoroborate Potassium p-tolyltrifluoroborate
57,612-3 C7H7BF3K 1g; 5g 57,155-5 C7H7BF3K 1g; 5g
576123
571555
Potassium 3-methoxyphenyltri-fluoroborate Potassium 3,5-bis(trifluoromethyl)-phenyltrifluoroborate
57,611-5 C7H7BF3KO 1g; 5g 57,156-3 C8H3BF9K 1g; 5g
576115
571563
Potassium trans-styryltrifluoroborate Potassium phenethyltrifluoroborate
57,615-8 C8H7BF3K 1g; 5g 56,309-9 C8H7BF3K 1g; 5g
576158
563099