Click Here! To visit the Chemical Synthesis Homepage.
Monthly Chemistry eNewsletter from Sigma-Aldrich
Theme: "Carbon-Carbon Bond Formation"
Hot News
Chem Product Central™
Click here to visit the chemistry focused weblog from Sigma-Aldrich.
NEW LITERATURE
Vol. 6, No. 7
Multi-Component Reactions
(1.12 MB PDF)
Vol. 6, No. 8
Privileged Ligands
(1.1 MB PDF)
Click here to subscribe to ChemFiles.
NEW PRODUCTS
Palladium Catalysts
674702
668532
668923
PEPPSI-SIPr is a novel N-Heterocyclic (NHC)-based catalyst that offers remarkable scope, reactivity, and stability in C-C and C-N bond-forming processes. Very active oxime-palladacycle catalyst for C-C bond-forming reactions ranging from the Suzuki to Ullman-type couplings in aqueous media. Highly robust palladacycle catalyst that has successfully mediated C-C bond-forming processes, ranging from the Heck to the Sonogashira to the Suzuki reaction in moderate to excellent yields.
Phosphines for Cross Coupling
675709
675938
The combination of this P,O-ligand with Pd(dba)2 generates a highly active catalyst system that exhibits broad utility in the Suzuki-Miyaura reaction. Bulky biaryl ligand that has proven highly useful for the unprecedented coupling of aryl chlorides and bromides with allylic alcohols via formal C-O bond formation.
676187
676179
Neopentyl-substituted phosphines form highly active catalysts in conjunction with Pd precursors for the amination of a wide range of amines with aryl halides.  The second ligand shown above has been successfully applied in both cross-coupling and amination reactions under aqueous conditions.
Boron Reagents for Suzuki Coupling
666491
666556
666459
673854
673862
652342
666793
674958
654396
676659
677787
677809
Organotin and Grignard Reagents
675660
673013
675679
Halogenated Substrates for Suzuki Coupling
652636
673994
669326
669350
669369
676063
676039
677736
666416
667366
Scavengers
668583
668605
668591
Other Catalysts
Other Reagents
667080
674613
Acts as an organocatalyst in highly enantioselective intramolecular Stetter reactions. Used in conjunction with CF3I for nucleophilic trifluoromethylation of aldehydes and ketones and chiral N-tolylsulfinimines.
PRODUCT SPOTLIGHTS
Click Chemistry: A "Click" Away from Drug Discovery.
Organotrifluoroborates: Potent Boronic Acid Surrogates.
Benzoylation Reagents: Simple tools for α-oxygenation of aldehydes and ketones.
Click Here for more product highlights.