|
Accelerate Methodology Discovery, Lead Generation, and Process Optimization
Aldrich’s new chemistry kits for catalysis screening offer the broadest range of technologies available in convenient packaging. Kits contain sets of catalysts, achiral or chiral ligands, and scavengers with varying steric and electronic properties that will accelerate your research discoveries. Kits offer several advantages:
- Enable rapid screening of catalytic processes for optimization
- Conveniently packaged in sampler format
- Cost of kit less than the total cost of individual components
New! Kits Containing New Aldrich Products
Buchwald Ligands I
Utilized for aromatic carbon-heteroatom formation and Suzuki coupling:
659932-1KT 1-g bottles |
NHC Ligands I
Sterically and electronically varied
to generate metal-based catalysts
with different properties:
662232-1KT 1-g bottles
|
|
BINOL Ligands I
BINOLs are privileged ligands that exhibit high levels of enantiocontrol in various catalytic transformations:
669113-1KT |
QuadraPureTM Scavengers
These macro- and microporous resins
effectively remove metal contamination
during chemical processing from R&D to manufacturing scale:
659428-1KT 5-g bottles |
|
Other Catalysis kits from Sigma-Aldrich |
|
Homogeneous Pd Catalysts I
PdCl2 · Pd(OAc)2 · Pd2(dba)3 · Pd(PPh3)2Cl2 · Pd(PPh3)4 · [Pd(allyl)Cl]2 · Pd(dppf)Cl2
|
659975-1KT 1-g bottles |
Heterogeneous Pd Catalysts I
· Palladium on Barium · Palladium on Calcium Carbonate · Palladium on Alumina · Palladium on Activated Carbon · Palladium (Dry) on Activated Carbon · Palladium Hydroxide
|
659967-1KT 1-g bottles |
Pd EnCatTM
Pd EnCatTM 30 · Pd EnCatTM 40 · Pd EnCatTM TPP30
· Pd EnCatTM TOTP30 · Pd EnCatTM BINAP30
| 666335-1KT 1-g bottles |
Phosphine Ligands I
PPh3 · PCy3 · P(t-Bu)3H-BF4 · P(o-tolyl)3 · P(2-Furyl)3 · dppp · dppf
| 659940-1KT 1-g bottles |
|